Product References
Mechanical Unloading Promotes Osteoclastic Differentiation and Bone Resorption by Modulating the MSC Secretome to Favor Inflammation.
Cell transplantation
Wang W,Zheng X,Wang H,Zuo B,Chen S,Li J
Published figure using CD11b/c monoclonal antibody (Product # 12-0110-82) in Flow Cytometry
Wed Mar 20 00:00:00 UTC 2024
Protective effect of HDACIs in improves survival and organ injury after CLP-induced sepsis.
Surgery open science
Niu K,Qu S,Yang L,Zhang H,Yuan J,Fan H,Li X,Tao K
12-0110-82 was used in Flow cytometry/Cell sorting to indicate that HDACI classes I and IIb and non-selective HDACI can significantly prolong survival.
Wed Mar 01 00:00:00 UTC 2023
Atractylenolide III ameliorates spinal cord injury in rats by modulating microglial/macrophage polarization.
CNS neuroscience & therapeutics
Xue MT,Sheng WJ,Song X,Shi YJ,Geng ZJ,Shen L,Wang R,Lü HZ,Hu JG
12-0110-82 was used in flow cytometry to reveal a novel effect of ATL-III in the regulation of microglial/macrophage polarization and provides initial evidence that ATL-III has potential therapeutic benefits in SCI rats.
Fri Jul 01 00:00:00 UTC 2022
Negative pressure wound therapy improves bone regeneration by promoting osteogenic differentiation via the AMPK-ULK1-autophagy axis.
Autophagy
Zhang S,Xie Y,Yan F,Zhang Y,Yang Z,Chen Z,Zhao Y,Huang Z,Cai L,Deng Z
12-0110-82 was used in Flow cytometry/Cell sorting to identify a novel AMPK-ULK1-autophagy axis by which negative pressure promotes osteoblast differentiation of MSCs and bone regeneration.
Thu Sep 01 00:00:00 UTC 2022
Effects of combination treatment with transcranial magnetic stimulation and bone marrow mesenchymal stem cell transplantation or Raf inhibition on spinal cord injury in rats.
Molecular medicine reports
Feng S,Wang S,Sun S,Su H,Zhang L
Published figure using CD11b/c monoclonal antibody (Product # 12-0110-82) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2021
Disruption of Tfh:B Cell Interactions Prevents Antibody-Mediated Rejection in a Kidney Transplant Model in Rats: Impact of Calcineurin Inhibitor Dose.
Frontiers in immunology
Steines L,Poth H,Schuster A,Amann K,Banas B,Bergler T
12-0110-82 was used in Flow Cytometry to conclude that Tfh:B cell crosstalk was controlled only by hiCNI treatment, preventing the development of DSA and ABMR.
Tue Sep 28 00:00:00 UTC 2021
Disruption of Tfh:B Cell Interactions Prevents Antibody-Mediated Rejection in a Kidney Transplant Model in Rats: Impact of Calcineurin Inhibitor Dose.
Frontiers in immunology
Steines L,Poth H,Schuster A,Amann K,Banas B,Bergler T
12-0110-82 was used in Flow Cytometry to conclude that Tfh:B cell crosstalk was controlled only by hiCNI treatment, preventing the development of DSA and ABMR.
Tue Sep 28 00:00:00 UTC 2021
Macrophage-Derived Inflammation Induces a Transcriptome Makeover in Mesenchymal Stromal Cells Enhancing Their Potential for Tissue Repair.
International journal of molecular sciences
Maldonado-Lasunción I,O'Neill N,Umland O,Verhaagen J,Oudega M
12-0110-82 was used in Flow cytometry/Cell sorting to investigate macrophage-derived inflammation as a means to condition mesenchymal stromal cell by comprehensively analyzing their transcriptome and secretome.
Thu Jan 14 00:00:00 UTC 2021
Placenta‑derived mesenchymal stem cells ameliorate lipopolysaccharide‑induced inflammation in RAW264.7 cells and acute lung injury in rats.
Molecular medicine reports
Yuan W,Song HY,Xiong J,Jiang WL,Kang GJ,Huang J,Xie SP
12-0110 was used in Flow cytometry/Cell sorting to demonstrate placenta‚derived mesenchymal stem cells could be used as therapy for lipopolysaccharide (LPS)‚induced acute lung injury.
Sat Aug 01 00:00:00 UTC 2020
Cardioprotective Effects of Latifolin Against Doxorubicin-Induced Cardiotoxicity by Macrophage Polarization in Mice.
Journal of cardiovascular pharmacology
Zhang N,Shou B,Chen L,Lai X,Luo Y,Meng X,Liu R
12-0110 was used in Flow cytometry/Cell sorting to demonstrate that latifolin prevented DOX-induced cardiac dysfunction and produced macrophage polarization in mice challenged with latifolin.
Mon Jun 01 00:00:00 UTC 2020
Bifidobacteria shape host neural circuits during postnatal development by promoting synapse formation and microglial function.
Scientific reports
Luck B,Engevik MA,Ganesh BP,Lackey EP,Lin T,Balderas M,Major A,Runge J,Luna RA,Sillitoe RV,Versalovic J
12-0110 was used in Flow cytometry/Cell sorting to indicate that neonatal microbial colonization of the gut elicits concomitant effects on the host CNS, which promote the homeostatic developmental balance of neural connections during the postnatal time period.
Fri May 08 00:00:00 UTC 2020
Bifidobacteria shape host neural circuits during postnatal development by promoting synapse formation and microglial function.
Scientific reports
Luck B,Engevik MA,Ganesh BP,Lackey EP,Lin T,Balderas M,Major A,Runge J,Luna RA,Sillitoe RV,Versalovic J
12-0110 was used in Flow cytometry/Cell sorting to indicate that neonatal microbial colonization of the gut elicits concomitant effects on the host CNS, which promote the homeostatic developmental balance of neural connections during the postnatal time period.
Fri May 08 00:00:00 UTC 2020
Sodium bicarbonate loading limits tubular cast formation independent of glomerular injury and proteinuria in Dahl salt-sensitive rats.
Clinical science (London, England : 1979)
Ray SC,Patel B,Irsik DL,Sun J,Ocasio H,Crislip GR,Jin CH,Chen J,Baban B,Polichnowski AJ,O'Connor PM
12-0110-82 was used in Flow Cytometry to study whether NaHCO3 protects from the development of renal injury in Dahl salt-sensitive rats via luminal alkalization which limits the formation of tubular casts, which are a prominent pathological feature in this model.
Fri Jun 15 00:00:00 UTC 2018
Genetic deletion of microglial Panx1 attenuates morphine withdrawal, but not analgesic tolerance or hyperalgesia in mice.
Channels (Austin, Tex.)
Burma NE,Leduc-Pessah H,Trang T
12-0110-82 was used in Flow Cytometry to investigate whether microglial Panx1 contributes to opioid-induced hyperalgesia and opioid analgesic tolerance using mice with a tamoxifen-inducible deletion of microglial Panx1.
Sun Sep 03 00:00:00 UTC 2017
Chitosan hydrogel improves mesenchymal stem cell transplant survival and cardiac function following myocardial infarction in rats.
Experimental and therapeutic medicine
Xu B,Li Y,Deng B,Liu X,Wang L,Zhu QL
12-0110 was used in Flow cytometry/Cell sorting to evaluate the suitability of chitosan hydrogel, an injectable scaffold, to deliver mesenchymal stem cells into infarcted myocardium.
Wed Feb 01 00:00:00 UTC 2017
Renal allograft rejection, lymphocyte infiltration, and de novo donor-specific antibodies in a novel model of non-adherence to immunosuppressive therapy.
BMC immunology
Kühne L,Jung B,Poth H,Schuster A,Wurm S,Ruemmele P,Banas B,Bergler T
12-0110 was used in Flow cytometry/Cell sorting to investigate the immunological mechanisms underlying chronic renal allograft rejection in a model of non-adherence to immunosuppressive therapy.
Tue Dec 19 00:00:00 UTC 2017
Persistent inflammatory state after photoreceptor loss in an animal model of retinal degeneration.
Scientific reports
Noailles A,Maneu V,Campello L,Gómez-Vicente V,Lax P,Cuenca N
12-0110 was used in Flow cytometry/Cell sorting to assess longitudinal changes in microglia during retinal degeneration in a model of dominant retinitis pigmentosa.
Wed Sep 14 00:00:00 UTC 2016
Leucine-rich repeat kinase 2 deficiency is protective in rhabdomyolysis-induced kidney injury.
Human molecular genetics
Boddu R,Hull TD,Bolisetty S,Hu X,Moehle MS,Daher JP,Kamal AI,Joseph R,George JF,Agarwal A,Curtis LM,West AB
12-0110 was used in Flow cytometry/Cell sorting to highlight several novel physiological roles for leucine-rich repeat kinase 2 in disposition to kidney disease.
Wed Jul 15 00:00:00 UTC 2015