Save $650 & get a lab-themed hidden object puzzle

Save and claim

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • CD138 (Syndecan-1) Antibodies

            Invitrogen

            CD138 (Syndecan-1) Monoclonal Antibody (DL-101), FITC, eBioscience™

            8 Published Figures
            10 References
            View all (91) CD138 (Syndecan-1) antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 11-1389-42
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            239.70
            Special offer
            Online exclusive
            282.00
            Save 42.30 (15%)
            100 Tests
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite CD138 (Syndecan-1) Monoclonal Antibody (DL-101), FITC, eBioscience™

            • Antibody Testing Data (1)
            • Published Figures (8)
            Application
            CD138 (Syndecan-1) Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 9

            CD138 (Syndecan-1) Antibody (11-1389-42) in Flow

            Staining of the U266 cell line with Mouse IgG1 K Isotype Control FITC (Product # 11-4714-42) (blue histogram) or Anti-Human CD138 (Syndecan-1) FITC (purple histogram). Total viable cells, as determined by Fixable Viability Dye eFluor® 660, were used for analysis.
            View Product

            CD138 (Syndecan-1) Antibody in Flow Cytometry (Flow)
            CD138 (Syndecan-1) Antibody in Flow Cytometry (Flow)
            CD138 (Syndecan-1) Antibody in Flow Cytometry (Flow)
            CD138 (Syndecan-1) Antibody in Flow Cytometry (Flow)
            CD138 (Syndecan-1) Antibody in Flow Cytometry (Flow)
            CD138 (Syndecan-1) Antibody in Flow Cytometry (Flow)
            CD138 (Syndecan-1) Antibody in Flow Cytometry (Flow)
            CD138 (Syndecan-1) Antibody in Flow Cytometry (Flow)
            CD138 (Syndecan-1) Antibody in Immunocytochemistry (ICC/IF)

            Product Details

            11-1389-42

            Applications
            Tested Dilution
            Publications

            Immunocytochemistry (ICC/IF)

            -
            View 1 publication 1 publication

            Flow Cytometry (Flow)

            5 µL (1 µg)/test
            View 9 publications 9 publications
            Product Specifications

            Species Reactivity

            Human

            Published species

            Human, Rhesus monkey

            Host/Isotype

            Mouse / IgG1, kappa

            Recommended Isotype Control

            Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), FITC, eBioscience™

            Class

            Monoclonal

            Type

            Antibody

            Clone

            DL-101

            Conjugate

            FITC
            • Unconjugated
            • APC 
            • PE 
            • PE-Cyanine7
            • Super Bright 436
            • Super Bright 600
            • Request custom conjugation

            Excitation/Emission Max

            498/517 nm View spectra spectra

            Form

            Liquid

            Concentration

            5 µL/Test

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2, with BSA

            Contains

            0.09% sodium azide

            Storage conditions

            4°C, store in dark, DO NOT FREEZE!

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_11039872

            Product Specific Information

            Description: The DL-101 monoclonal antibody reacts with human CD138, also known as syndecan-1. CD138 is a transmembrane protein containing chondroitin sulfate and heparin sulfate moieties responsible for binding to the extracellular matrix components. CD138 is not expressed by mature B cells but is present on pre-B cells and the finally differentiated plasma cells.

            Applications Reported: DL-101 has been reported for use in flow cytometric analysis.

            Applications Tested: This DL-101 antibody has been pre-titrated and tested by flow cytometric analysis of the U266 cell line. This can be used at 5 µL (1 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

            Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            CD138 (syndecan 1) is a transmembrane proteoglycan that can bind a variety of cytokines and modulate their activity, as well as the activity of extracellular matrix components and influence many developmental processes. CD138 is expressed mainly in differentiating keratinocytes and is transiently upregulated in all layers of the epidermis upon tissue injury. It is also highly expressed on plasma cells and can be detected even on fibroblasts, vascular smooth muscle cells and endothelial cells. Up-regulation and down-regulation of CD138 on the cell surface often correlates with the gain of cancerous characteristics. Serum levels of the shedded soluble sCD138 are used as a prognostic factor of cancerogenesis.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            How to build a flow cytometry experiment with Invitrogen Flow Cytometry Panel Builder
            The Invitrogen Flow Cytometry Panel Builder webtool helps you chose antibody clones, fluorophores, fluorescent proteins, and reagents to design your experiment.
            Video Player is loading.
            Current Time 0:00
            /
            Duration 8:21
            Loaded: 0.00%
            0:00
            Stream Type LIVE
            Remaining Time -8:21
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected
            • en (Main), selected

            This is a modal window.

            Beginning of dialog window. Escape will cancel and close the window.

            Text
            Text Background
            Caption Area Background
            Font Size
            Text Edge Style
            Font Family

            End of dialog window.

            This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

            How to use the Panel Builder

            Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

            Bioinformatics

            Protein Aliases: CD antigen 138; CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; sCD138; soluble CD138; SYND1; syndecan proteoglycan 1; Syndecan-1; Syndecan1

            View more View less

            Gene Aliases: CD138; SDC; SDC1; SYND1; syndecan

            View more View less

            UniProt ID: (Human) P18827

            View more View less

            Entrez Gene ID: (Human) 6382

            View more View less

            Function(s)

            glycoprotein binding protein binding protein C-terminus binding transmembrane signal receptor

            Process(es)

            retinoid metabolic process ureteric bud development glycosaminoglycan biosynthetic process glycosaminoglycan catabolic process inflammatory response response to toxic substance cell migration glycosaminoglycan metabolic process wound healing lipoprotein metabolic process odontogenesis response to hydrogen peroxide myoblast development response to glucocorticoid response to cAMP response to calcium ion striated muscle cell development Sertoli cell development canonical Wnt signaling pathway positive regulation of exosomal secretion positive regulation of extracellular exosome assembly
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-kvfvs:80/100.66.77.138:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0