Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MERASCLLLL LLPLVHVSAT TPEPCELDDE DFRCVCNFSE PQPDWSEAFQ CVSAVEVEIH AGGLNLEPFL KRVDADADPR QYADTVKALR VRRLTVGAAQ VPAQLLVGAL RVLAYSRLKE LTLEDLKITG TMPPLPLEAT GLALSSLRLR NVSWATGRSW LAELQQWLKP GLKVLSIAQA HSPAFSCEQV RAFPALTSLD LSDNPGLGER GLMAALCPHK FPAIQNLALR NTGMETPTGV CAALAAAGVQ PHSLDLSHNS LRATVNPSAP RCMWSSALNS LNLSFAGLEQ VPKGLPAKLR VLDLSCNRLN RAPQPDELPE VDNLTLDGNP FLVPGTALPH EGSMNSGVVP ACARSTLSVG VSGTLVLLQG ARGFA
CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD 14; CD14; cd14 monocyte; LPSR antibody; Monocyte differentiation antigen CD14; My23 antigen; Myeloid cell-specific leucine-rich glycoprotein; sCD14; soluble CD14
Gene Aliases: CD14
UniProt ID: (Human) P08571
Entrez Gene ID: (Human) 929
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support