Product References
Distinct niche structures and intrinsic programs of fallopian tube and ovarian surface epithelial cells.
iScience
Qin G,Park ES,Chen X,Han S,Xiang D,Ren F,Liu G,Chen H,Yuan GC,Li Z
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Fri Jan 20 00:00:00 EST 2023
The Wnt/TCF7L1 transcriptional repressor axis drives primitive endoderm formation by antagonizing naive and formative pluripotency.
Nature communications
Athanasouli P,Balli M,De Jaime-Soguero A,Boel A,Papanikolaou S,van der Veer BK,Janiszewski A,Vanhessche T,Francis A,El Laithy Y,Nigro AL,Aulicino F,Koh KP,Pasque V,Cosma MP,Verfaillie C,Zwijsen A,Heindryckx B,Nikolaou C,Lluis F
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Fri Mar 03 00:00:00 EST 2023
An estrogen-sensitive fibroblast population drives abdominal muscle fibrosis in an inguinal hernia mouse model.
JCI insight
Potluri T,Taylor MJ,Stulberg JJ,Lieber RL,Zhao H,Bulun SE
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Apr 19 00:00:00 EDT 2022
Hematopoietic-Mesenchymal Signals Regulate the Properties of Mesenchymal Stem Cells.
International journal of molecular sciences
Kanazawa S,Okada H,Riu D,Mabuchi Y,Akazawa C,Iwata J,Hoshi K,Hikita A
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Jul 26 00:00:00 EDT 2022
PEAR1 regulates expansion of activated fibroblasts and deposition of extracellular matrix in pulmonary fibrosis.
Nature communications
Geng Y,Li L,Yan J,Liu K,Yang A,Zhang L,Shen Y,Gao H,Wu X,Noth I,Huang Y,Liu J,Fan X
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Sat Nov 19 00:00:00 EST 2022
Lamin A/C-dependent chromatin architecture safeguards naïve pluripotency to prevent aberrant cardiovascular cell fate and function.
Nature communications
Wang Y,Elsherbiny A,Kessler L,Cordero J,Shi H,Serke H,Lityagina O,Trogisch FA,Mohammadi MM,El-Battrawy I,Backs J,Wieland T,Heineke J,Dobreva G
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Fri Nov 04 00:00:00 EDT 2022
Placental endocrine function shapes cerebellar development and social behavior.
Nature neuroscience
Vacher CM,Lacaille H,O'Reilly JJ,Salzbank J,Bakalar D,Sebaoui S,Liere P,Clarkson-Paredes C,Sasaki T,Sathyanesan A,Kratimenos P,Ellegood J,Lerch JP,Imamura Y,Popratiloff A,Hashimoto-Torii K,Gallo V,Schumacher M,Penn AA
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunohistochemistry
Fri Oct 01 00:00:00 EDT 2021
IGF1R controls mechanosignaling in myofibroblasts required for pulmonary alveologenesis.
JCI insight
He H,Snowball J,Sun F,Na CL,Whitsett JA
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Mon Mar 22 00:00:00 EDT 2021
Interleukin-11-expressing fibroblasts have a unique gene signature correlated with poor prognosis of colorectal cancer.
Nature communications
Nishina T,Deguchi Y,Ohshima D,Takeda W,Ohtsuka M,Shichino S,Ueha S,Yamazaki S,Kawauchi M,Nakamura E,Nishiyama C,Kojima Y,Adachi-Akahane S,Hasegawa M,Nakayama M,Oshima M,Yagita H,Shibuya K,Mikami T,Inohara N,Matsushima K,Tada N,Nakano H
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Fri Apr 16 00:00:00 EDT 2021
Zbtb16 regulates social cognitive behaviors and neocortical development.
Translational psychiatry
Usui N,Berto S,Konishi A,Kondo M,Konopka G,Matsuzaki H,Shimada S
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunohistochemistry
Sat Apr 24 00:00:00 EDT 2021
Gain-of-function p53 protein transferred via small extracellular vesicles promotes conversion of fibroblasts to a cancer-associated phenotype.
Cell reports
Ma S,McGuire MH,Mangala LS,Lee S,Stur E,Hu W,Bayraktar E,Villar-Prados A,Ivan C,Wu SY,Yokoi A,Dasari SK,Jennings NB,Liu J,Lopez-Berestein G,Ram P,Sood AK
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Feb 09 00:00:00 EST 2021
Dlx5-augmentation in neural crest cells reveals early development and differentiation potential of mouse apical head mesenchyme.
Scientific reports
Vu TH,Takechi M,Shimizu M,Kitazawa T,Higashiyama H,Iwase A,Kurihara H,Iseki S
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunohistochemistry
Fri Jan 22 00:00:00 EST 2021
Extracellular vesicles synchronize cellular phenotypes of differentiating cells.
Journal of extracellular vesicles
Minakawa T,Matoba T,Ishidate F,Fujiwara TK,Takehana S,Tabata Y,Yamashita JK
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunocytochemistry
Wed Sep 01 00:00:00 EDT 2021
Extracellular vesicles synchronize cellular phenotypes of differentiating cells.
Journal of extracellular vesicles
Minakawa T,Matoba T,Ishidate F,Fujiwara TK,Takehana S,Tabata Y,Yamashita JK
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunocytochemistry
Wed Sep 01 00:00:00 EDT 2021
Mesothelial cells are not a source of adipocytes in mice.
Cell reports
Westcott GP,Emont MP,Li J,Jacobs C,Tsai L,Rosen ED
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Jul 13 00:00:00 EDT 2021
Mesothelial cells are not a source of adipocytes in mice.
Cell reports
Westcott GP,Emont MP,Li J,Jacobs C,Tsai L,Rosen ED
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Jul 13 00:00:00 EDT 2021
Epithelial Vegfa Specifies a Distinct Endothelial Population in the Mouse Lung.
Developmental cell
Vila Ellis L,Cain MP,Hutchison V,Flodby P,Crandall ED,Borok Z,Zhou B,Ostrin EJ,Wythe JD,Chen J
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunocytochemistry
Mon Mar 09 00:00:00 EDT 2020
A smooth muscle-derived, Braf-driven mouse model of gastrointestinal stromal tumor (GIST): evidence for an alternative GIST cell-of-origin.
The Journal of pathology
Kondo J,Huh WJ,Franklin JL,Heinrich MC,Rubin BP,Coffey RJ
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunocytochemistry
Tue Dec 01 00:00:00 EST 2020
Hyaluronan Degradation by Cemip Regulates Host Defense against Staphylococcus aureus Skin Infection.
Cell reports
Dokoshi T,Zhang LJ,Li F,Nakatsuji T,Butcher A,Yoshida H,Shimoda M,Okada Y,Gallo RL
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Jan 07 00:00:00 EST 2020
Tissue-Resident PDGFRα+ Progenitor Cells Contribute to Fibrosis versus Healing in a Context- and Spatiotemporally Dependent Manner.
Cell reports
Santini MP,Malide D,Hoffman G,Pandey G,D'Escamard V,Nomura-Kitabayashi A,Rovira I,Kataoka H,Ochando J,Harvey RP,Finkel T,Kovacic JC
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Jan 14 00:00:00 EST 2020
Loss of Fas signaling in fibroblasts impairs homeostatic fibrosis resolution and promotes persistent pulmonary fibrosis.
JCI insight
Redente EF,Chakraborty S,Sajuthi S,Black BP,Edelman BL,Seibold MA,Riches DW
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Dec 08 00:00:00 EST 2020
Adipocyte Piezo1 mediates obesogenic adipogenesis through the FGF1/FGFR1 signaling pathway in mice.
Nature communications
Wang S,Cao S,Arhatte M,Li D,Shi Y,Kurz S,Hu J,Wang L,Shao J,Atzberger A,Wang Z,Wang C,Zang W,Fleming I,Wettschureck N,Honoré E,Offermanns S
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Fri May 08 00:00:00 EDT 2020
Bim expression modulates the pro-inflammatory phenotype of retinal astroglial cells.
PloS one
Falero-Perez J,Sheibani N,Sorenson CM
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Fri Jul 31 00:00:00 EDT 2020
Cardiac fibroblast proliferation rates and collagen expression mature early and are unaltered with advancing age.
JCI insight
Wu R,Ma F,Tosevska A,Farrell C,Pellegrini M,Deb A
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Thu Dec 17 00:00:00 EST 2020
Phosphatase SHP1 impedes mesenchymal stromal cell immunosuppressive capacity modulated by JAK1/STAT3 and P38 signals.
Cell & bioscience
Jiang M,Ye J,Wang X,Li N,Wang Y,Shi Y
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Thu Apr 14 00:00:00 EDT 2022
Interstitial cells of Cajal in Wsh/Wsh c-kit mutant mice.
Journal of smooth muscle research = Nihon Heikatsukin Gakkai kikanshi
Iino S,Horiguchi S,Horiguchi K,Hashimoto T
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunohistochemistry
Fri Jul 16 00:00:00 EDT 2021
Expression Profiling of Fibroblasts in Chronic and Acute Disease Models Reveals Novel Pathways in Kidney Fibrosis.
Journal of the American Society of Nephrology : JASN
Higashi AY,Aronow BJ,Dressler GR
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunohistochemistry
Tue Jan 01 00:00:00 EST 2019
PDGFRA defines the mesenchymal stem cell Kaposi's sarcoma progenitors by enabling KSHV oncogenesis in an angiogenic environment.
PLoS pathogens
Naipauer J,Rosario S,Gupta S,Premer C,Méndez-Solís O,Schlesinger M,Ponzinibbio V,Jain V,Gay L,Renne R,Chan HL,Morey L,Salyakina D,Abba M,Williams S,Hare JM,Goldschmidt-Clermont PJ,Mesri EA
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Sun Dec 01 00:00:00 EST 2019
Dual lineage tracing identifies intermediate mesenchymal stage for endocardial contribution to fibroblasts, coronary mural cells, and adipocytes.
The Journal of biological chemistry
Huang X,Feng T,Jiang Z,Meng J,Kou S,Lu Z,Chen W,Lin CP,Zhou B,Zhang H
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Fri May 31 00:00:00 EDT 2019
Growth Factor Screening in Dystrophic Muscles Reveals PDGFB/PDGFRB-Mediated Migration of Interstitial Stem Cells.
International journal of molecular sciences
Camps J,Grosemans H,Gijsbers R,Maes C,Sampaolesi M
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Mar 05 00:00:00 EST 2019
Adrenergic nerve degeneration in bone marrow drives aging of the hematopoietic stem cell niche.
Nature medicine
Maryanovich M,Zahalka AH,Pierce H,Pinho S,Nakahara F,Asada N,Wei Q,Wang X,Ciero P,Xu J,Leftin A,Frenette PS
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Bone marrow-derived fibroblasts are a functionally distinct stromal cell population in breast cancer.
The Journal of experimental medicine
Raz Y,Cohen N,Shani O,Bell RE,Novitskiy SV,Abramovitz L,Levy C,Milyavsky M,Leider-Trejo L,Moses HL,Grisaru D,Erez N
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunohistochemistry
Mon Dec 03 00:00:00 EST 2018
SCL/TAL1 cooperates with Polycomb RYBP-PRC1 to suppress alternative lineages in blood-fated cells.
Nature communications
Chagraoui H,Kristiansen MS,Ruiz JP,Serra-Barros A,Richter J,Hall-Ponselé E,Gray N,Waithe D,Clark K,Hublitz P,Repapi E,Otto G,Sopp P,Taylor S,Thongjuea S,Vyas P,Porcher C
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Dec 18 00:00:00 EST 2018
FZD4 Marks Lateral Plate Mesoderm and Signals with NORRIN to Increase Cardiomyocyte Induction from Pluripotent Stem Cell-Derived Cardiac Progenitors.
Stem cell reports
Yoon C,Song H,Yin T,Bausch-Fluck D,Frei AP,Kattman S,Dubois N,Witty AD,Hewel JA,Guo H,Emili A,Wollscheid B,Keller G,Zandstra PW
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Jan 09 00:00:00 EST 2018
Pdgfra marks a cellular lineage with distinct contributions to myofibroblasts in lung maturation and injury response.
eLife
Li R,Bernau K,Sandbo N,Gu J,Preissl S,Sun X
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Sep 04 00:00:00 EDT 2018
Age-specific biological and molecular profiling distinguishes paediatric from adult acute myeloid leukaemias.
Nature communications
Chaudhury S,O'Connor C,Cañete A,Bittencourt-Silvestre J,Sarrou E,Prendergast Á,Choi J,Johnston P,Wells CA,Gibson B,Keeshan K
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Dec 11 00:00:00 EST 2018
Genome editing reveals a role for OCT4 in human embryogenesis.
Nature
Fogarty NME,McCarthy A,Snijders KE,Powell BE,Kubikova N,Blakeley P,Lea R,Elder K,Wamaitha SE,Kim D,Maciulyte V,Kleinjung J,Kim JS,Wells D,Vallier L,Bertero A,Turner JMA,Niakan KK
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Thu Oct 05 00:00:00 EDT 2017
Preexisting endothelial cells mediate cardiac neovascularization after injury.
The Journal of clinical investigation
He L,Huang X,Kanisicak O,Li Y,Wang Y,Li Y,Pu W,Liu Q,Zhang H,Tian X,Zhao H,Liu X,Zhang S,Nie Y,Hu S,Miao X,Wang QD,Wang F,Chen T,Xu Q,Lui KO,Molkentin JD,Zhou B
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Tue Aug 01 00:00:00 EDT 2017
Attenuated PDGF signaling drives alveolar and microvascular defects in neonatal chronic lung disease.
EMBO molecular medicine
Oak P,Pritzke T,Thiel I,Koschlig M,Mous DS,Windhorst A,Jain N,Eickelberg O,Foerster K,Schulze A,Goepel W,Reicherzer T,Ehrhardt H,Rottier RJ,Ahnert P,Gortner L,Desai TJ,Hilgendorff A
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunocytochemistry
Wed Nov 01 00:00:00 EDT 2017
Intermittent Glucocorticoid Dosing Improves Muscle Repair and Function in Mice with Limb-Girdle Muscular Dystrophy.
The American journal of pathology
Quattrocelli M,Salamone IM,Page PG,Warner JL,Demonbreun AR,McNally EM
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunocytochemistry
Wed Nov 01 00:00:00 EDT 2017
Gli1 identifies osteogenic progenitors for bone formation and fracture repair.
Nature communications
Shi Y,He G,Lee WC,McKenzie JA,Silva MJ,Long F
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Mon Dec 11 00:00:00 EST 2017
Cardiac Fibro-Adipocyte Progenitors Express Desmosome Proteins and Preferentially Differentiate to Adipocytes Upon Deletion of the Desmoplakin Gene.
Circulation research
Lombardi R,Chen SN,Ruggiero A,Gurha P,Czernuszewicz GZ,Willerson JT,Marian AJ
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Fri Jun 24 00:00:00 EDT 2016
Developmental regression of hyaloid vasculature is triggered by neurons.
The Journal of experimental medicine
Yoshikawa Y,Yamada T,Tai-Nagara I,Okabe K,Kitagawa Y,Ema M,Kubota Y
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunohistochemistry
Mon Jun 27 00:00:00 EDT 2016
Developmental regression of hyaloid vasculature is triggered by neurons.
The Journal of experimental medicine
Yoshikawa Y,Yamada T,Tai-Nagara I,Okabe K,Kitagawa Y,Ema M,Kubota Y
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunohistochemistry
Mon Jun 27 00:00:00 EDT 2016
Pericyte-fibroblast transition promotes tumor growth and metastasis.
Proceedings of the National Academy of Sciences of the United States of America
Hosaka K,Yang Y,Seki T,Fischer C,Dubey O,Fredlund E,Hartman J,Religa P,Morikawa H,Ishii Y,Sasahara M,Larsson O,Cossu G,Cao R,Lim S,Cao Y
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Tue Sep 20 00:00:00 EDT 2016
The PDGF-BB-SOX7 axis-modulated IL-33 in pericytes and stromal cells promotes metastasis through tumour-associated macrophages.
Nature communications
Yang Y,Andersson P,Hosaka K,Zhang Y,Cao R,Iwamoto H,Yang X,Nakamura M,Wang J,Zhuang R,Morikawa H,Xue Y,Braun H,Beyaert R,Samani N,Nakae S,Hams E,Dissing S,Fallon PG,Langer R,Cao Y
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Fri May 06 00:00:00 EDT 2016
Genetic lineage tracing defines myofibroblast origin and function in the injured heart.
Nature communications
Kanisicak O,Khalil H,Ivey MJ,Karch J,Maliken BD,Correll RN,Brody MJ,J Lin SC,Aronow BJ,Tallquist MD,Molkentin JD
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Fri Jul 22 00:00:00 EDT 2016
Endothelial cells are progenitors of cardiac pericytes and vascular smooth muscle cells.
Nature communications
Chen Q,Zhang H,Liu Y,Adams S,Eilken H,Stehling M,Corada M,Dejana E,Zhou B,Adams RH
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Fri Aug 12 00:00:00 EDT 2016
Endothelial cells are progenitors of cardiac pericytes and vascular smooth muscle cells.
Nature communications
Chen Q,Zhang H,Liu Y,Adams S,Eilken H,Stehling M,Corada M,Dejana E,Zhou B,Adams RH
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Fri Aug 12 00:00:00 EDT 2016
mTOR inhibition and BMP signaling act synergistically to reduce muscle fibrosis and improve myofiber regeneration.
JCI insight
Agarwal S,Cholok D,Loder S,Li J,Breuler C,Chung MT,Sung HH,Ranganathan K,Habbouche J,Drake J,Peterson J,Priest C,Li S,Mishina Y,Levi B
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Thu Dec 08 00:00:00 EST 2016
mTOR inhibition and BMP signaling act synergistically to reduce muscle fibrosis and improve myofiber regeneration.
JCI insight
Agarwal S,Cholok D,Loder S,Li J,Breuler C,Chung MT,Sung HH,Ranganathan K,Habbouche J,Drake J,Peterson J,Priest C,Li S,Mishina Y,Levi B
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Thu Dec 08 00:00:00 EST 2016
Drug-based modulation of endogenous stem cells promotes functional remyelination in vivo.
Nature
Najm FJ,Madhavan M,Zaremba A,Shick E,Karl RT,Factor DC,Miller TE,Nevin ZS,Kantor C,Sargent A,Quick KL,Schlatzer DM,Tang H,Papoian R,Brimacombe KR,Shen M,Boxer MB,Jadhav A,Robinson AP,Podojil JR,Miller SD,Miller RH,Tesar PJ
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Thu Jun 11 00:00:00 EDT 2015
LRIG1 Regulates Ontogeny of Smooth Muscle-Derived Subsets of Interstitial Cells of Cajal in Mice.
Gastroenterology
Kondo J,Powell AE,Wang Y,Musser MA,Southard-Smith EM,Franklin JL,Coffey RJ
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Sat Aug 01 00:00:00 EDT 2015
The Isl1/Ldb1 Complex Orchestrates Genome-wide Chromatin Organization to Instruct Differentiation of Multipotent Cardiac Progenitors.
Cell stem cell
Caputo L,Witzel HR,Kolovos P,Cheedipudi S,Looso M,Mylona A,van IJcken WF,Laugwitz KL,Evans SM,Braun T,Soler E,Grosveld F,Dobreva G
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Thu Sep 03 00:00:00 EDT 2015
Mesodermal iPSC-derived progenitor cells functionally regenerate cardiac and skeletal muscle.
The Journal of clinical investigation
Quattrocelli M,Swinnen M,Giacomazzi G,Camps J,Barthélemy I,Ceccarelli G,Caluwé E,Grosemans H,Thorrez L,Pelizzo G,Muijtjens M,Verfaillie CM,Blot S,Janssens S,Sampaolesi M
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2015
IKKβ acts as a tumor suppressor in cancer-associated fibroblasts during intestinal tumorigenesis.
The Journal of experimental medicine
Pallangyo CK,Ziegler PK,Greten FR
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Mon Dec 14 00:00:00 EST 2015
Brg1 modulates enhancer activation in mesoderm lineage commitment.
Development (Cambridge, England)
Alexander JM,Hota SK,He D,Thomas S,Ho L,Pennacchio LA,Bruneau BG
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Wed Apr 15 00:00:00 EDT 2015
Oligodendroglial maturation is dependent on intracellular protein shuttling.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Göttle P,Sabo JK,Heinen A,Venables G,Torres K,Tzekova N,Parras CM,Kremer D,Hartung HP,Cate HS,Küry P
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Wed Jan 21 00:00:00 EST 2015
Activation of non-myogenic mesenchymal stem cells during the disease progression in dystrophic dystrophin/utrophin knockout mice.
Human molecular genetics
Sohn J,Lu A,Tang Y,Wang B,Huard J
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Wed Jul 01 00:00:00 EDT 2015
Activation of non-myogenic mesenchymal stem cells during the disease progression in dystrophic dystrophin/utrophin knockout mice.
Human molecular genetics
Sohn J,Lu A,Tang Y,Wang B,Huard J
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Wed Jul 01 00:00:00 EDT 2015
Alpha-Synuclein Expression in the Oligodendrocyte Lineage: an In Vitro and In Vivo Study Using Rodent and Human Models.
Stem cell reports
Djelloul M,Holmqvist S,Boza-Serrano A,Azevedo C,Yeung MS,Goldwurm S,Frisén J,Deierborg T,Roybon L
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Aug 11 00:00:00 EDT 2015
Cardiogenic genes expressed in cardiac fibroblasts contribute to heart development and repair.
Circulation research
Furtado MB,Costa MW,Pranoto EA,Salimova E,Pinto AR,Lam NT,Park A,Snider P,Chandran A,Harvey RP,Boyd R,Conway SJ,Pearson J,Kaye DM,Rosenthal NA
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Fri Apr 25 00:00:00 EDT 2014
Recruitment of bone marrow-derived cells to periodontal tissue defects.
Frontiers in cell and developmental biology
Kimura Y,Komaki M,Iwasaki K,Sata M,Izumi Y,Morita I
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Mon Nov 03 00:00:00 EST 2014
Cell-cycle dependent expression of a translocation-mediated fusion oncogene mediates checkpoint adaptation in rhabdomyosarcoma.
PLoS genetics
Kikuchi K,Hettmer S,Aslam MI,Michalek JE,Laub W,Wilky BA,Loeb DM,Rubin BP,Wagers AJ,Keller C
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Western Blot
Wed Jan 01 00:00:00 EST 2014
Distinct fibroblast lineages determine dermal architecture in skin development and repair.
Nature
Driskell RR,Lichtenberger BM,Hoste E,Kretzschmar K,Simons BD,Charalambous M,Ferron SR,Herault Y,Pavlovic G,Ferguson-Smith AC,Watt FM
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Thu Dec 12 00:00:00 EST 2013
Dynamic and nuclear expression of PDGFRα and IGF-1R in alveolar Rhabdomyosarcoma.
Molecular cancer research : MCR
Aslam MI,Hettmer S,Abraham J,Latocha D,Soundararajan A,Huang ET,Goros MW,Michalek JE,Wang S,Mansoor A,Druker BJ,Wagers AJ,Tyner JW,Keller C
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Western Blot
Fri Nov 01 00:00:00 EDT 2013
Dynamic and nuclear expression of PDGFRα and IGF-1R in alveolar Rhabdomyosarcoma.
Molecular cancer research : MCR
Aslam MI,Hettmer S,Abraham J,Latocha D,Soundararajan A,Huang ET,Goros MW,Michalek JE,Wang S,Mansoor A,Druker BJ,Wagers AJ,Tyner JW,Keller C
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Western Blot
Fri Nov 01 00:00:00 EDT 2013
Totipotent embryonic stem cells arise in ground-state culture conditions.
Cell reports
Morgani SM,Canham MA,Nichols J,Sharov AA,Migueles RP,Ko MS,Brickman JM
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Thu Jun 27 00:00:00 EDT 2013
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2012
Mosaic analysis with double markers reveals tumor cell of origin in glioma.
Cell
Liu C,Sage JC,Miller MR,Verhaak RG,Hippenmeyer S,Vogel H,Foreman O,Bronson RT,Nishiyama A,Luo L,Zong H
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Fri Jul 22 00:00:00 EDT 2011
ER71 directs mesodermal fate decisions during embryogenesis.
Development (Cambridge, England)
Rasmussen TL,Kweon J,Diekmann MA,Belema-Bedada F,Song Q,Bowlin K,Shi X,Ferdous A,Li T,Kyba M,Metzger JM,Koyano-Nakagawa N,Garry DJ
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2011
ER71 directs mesodermal fate decisions during embryogenesis.
Development (Cambridge, England)
Rasmussen TL,Kweon J,Diekmann MA,Belema-Bedada F,Song Q,Bowlin K,Shi X,Ferdous A,Li T,Kyba M,Metzger JM,Koyano-Nakagawa N,Garry DJ
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2011
Wt1 controls retinoic acid signalling in embryonic epicardium through transcriptional activation of Raldh2.
Development (Cambridge, England)
Guadix JA,Ruiz-Villalba A,Lettice L,Velecela V,Muñoz-Chápuli R,Hastie ND,Pérez-Pomares JM,Martínez-Estrada OM
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunofluorescence
Tue Mar 01 00:00:00 EST 2011
A functional role for the 'fibroblast-like cells' in gastrointestinal smooth muscles.
The Journal of physiology
Kurahashi M,Zheng H,Dwyer L,Ward SM,Koh SD,Sanders KM
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Immunocytochemistry
Tue Feb 01 00:00:00 EST 2011
CCAAT/enhancer binding protein beta regulates stem cell activity and specifies luminal cell fate in the mammary gland.
Stem cells (Dayton, Ohio)
LaMarca HL,Visbal AP,Creighton CJ,Liu H,Zhang Y,Behbod F,Rosen JM
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Wed Mar 31 00:00:00 EDT 2010
Bioluminescence imaging of Olig2-neural stem cells reveals improved engraftment in a demyelination mouse model.
Stem cells (Dayton, Ohio)
Sher F,van Dam G,Boddeke E,Copray S
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 11-1401-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2009
Olig2+ neuroepithelial motoneuron progenitors are not multipotent stem cells in vivo.
Proceedings of the National Academy of Sciences of the United States of America
Mukouyama YS,Deneen B,Lukaszewicz A,Novitch BG,Wichterle H,Jessell TM,Anderson DJ
Tue Jan 31 00:00:00 EST 2006
Tlx, an orphan nuclear receptor, regulates cell numbers and astrocyte development in the developing retina.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Miyawaki T,Uemura A,Dezawa M,Yu RT,Ide C,Nishikawa S,Honda Y,Tanabe Y,Tanabe T
Wed Sep 15 00:00:00 EDT 2004
PDGFR alpha expression during mouse embryogenesis: immunolocalization analyzed by whole-mount immunohistostaining using the monoclonal anti-mouse PDGFR alpha antibody APA5.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Takakura N,Yoshida H,Ogura Y,Kataoka H,Nishikawa S,Nishikawa S
Sun Jun 01 00:00:00 EDT 1997