Product References
Type H vessel/platelet-derived growth factor receptor β+ perivascular cell disintegration is involved in vascular injury and bone loss in radiation-induced bone damage.
Cell proliferation
Li J,Chen X,Ren L,Chen X,Wu T,Wang Y,Ren X,Cheng B,Xia J
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Sat Jul 01 00:00:00 EDT 2023
Release of stem cells from quiescence reveals gliogenic domains in the adult mouse brain.
Science (New York, N.Y.)
Delgado AC,Maldonado-Soto AR,Silva-Vargas V,Mizrak D,von Känel T,Tan KR,Paul A,Madar A,Cuervo H,Kitajewski J,Lin CS,Doetsch F
13-1402-82 was used in Flow Cytometry to show that platelet-derived growth factor receptor beta (PDGFRβ) is expressed by adult ventricular-subventricular zone neural stem cells that generate olfactory bulb interneurons and glia.
Fri Jun 11 00:00:00 EDT 2021
Cellular dormancy in minimal residual disease following targeted therapy.
Breast cancer research : BCR
Ruth JR,Pant DK,Pan TC,Seidel HE,Baksh SC,Keister BA,Singh R,Sterner CJ,Bakewell SJ,Moody SE,Belka GK,Chodosh LA
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Fri Jun 04 00:00:00 EDT 2021
Distinct Fibroblast Lineages Give Rise to NG2+ Pericyte Populations in Mouse Skin Development and Repair.
Frontiers in cell and developmental biology
Goss G,Rognoni E,Salameti V,Watt FM
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Wed Mar 02 00:00:00 EST 2022
Generation and characterization of a Myh6-driven Cre knockin mouse line.
Transgenic research
Huang X,Yan L,Kou S,Meng J,Lu Z,Lin CP,Liu C,Zhang H
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunocytochemistry
Wed Dec 01 00:00:00 EST 2021
Direct contribution of skeletal muscle mesenchymal progenitors to bone repair.
Nature communications
Julien A,Kanagalingam A,Martínez-Sarrà E,Megret J,Luka M,Ménager M,Relaix F,Colnot C
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunohistochemistry
Mon May 17 00:00:00 EDT 2021
Perivascular cell-derived extracellular vesicles stimulate colorectal cancer revascularization after withdrawal of antiangiogenic drugs.
Journal of extracellular vesicles
Huang M,Chen M,Qi M,Ye G,Pan J,Shi C,Yang Y,Zhao L,Mo X,Zhang Y,Li Y,Zhong J,Lu W,Li X,Zhang J,Lin J,Luo L,Liu T,Tang PM,Hong A,Cao Y,Ye W,Zhang D
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunocytochemistry
Sat May 01 00:00:00 EDT 2021
Cardiac pericytes function as key vasoactive cells to regulate homeostasis and disease.
FEBS open bio
Lee LL,Khakoo AY,Chintalgattu V
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
Single-cell profiling reveals an endothelium-mediated immunomodulatory pathway in the eye choroid.
The Journal of experimental medicine
Lehmann GL,Hanke-Gogokhia C,Hu Y,Bareja R,Salfati Z,Ginsberg M,Nolan DJ,Mendez-Huergo SP,Dalotto-Moreno T,Wojcinski A,Ochoa F,Zeng S,Cerliani JP,Panagis L,Zager PJ,Mullins RF,Ogura S,Lutty GA,Bang J,Zippin JH,Romano C,Rabinovich GA,Elemento O,Joyner AL,Rafii S,Rodriguez-Boulan E,Benedicto I
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunocytochemistry
Mon Jun 01 00:00:00 EDT 2020
Enhancement of Drug Seeking Following Drug Taking in a Sexual Context Requires Anterior Cingulate Cortex Activity in Male Rats.
Frontiers in behavioral neuroscience
Kuiper LB,Lucas KA,Mai V,Coolen LM
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunohistochemistry
Mon Sep 28 00:00:00 EDT 2020
TGFβ1 orchestrates renal fibrosis following Escherichia coli pyelonephritis.
Physiological reports
Hreha TN,Collins CA,Daugherty AL,Twentyman J,Paluri N,Hunstad DA
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunohistochemistry
Sun Mar 01 00:00:00 EST 2020
Angiomodulin (IGFBP7) is a cerebral specific angiocrine factor, but is probably not a blood-brain barrier inducer.
Fluids and barriers of the CNS
Bar O,Gelb S,Atamny K,Anzi S,Ben-Zvi A
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunocytochemistry
Wed Apr 01 00:00:00 EDT 2020
Angiomodulin (IGFBP7) is a cerebral specific angiocrine factor, but is probably not a blood-brain barrier inducer.
Fluids and barriers of the CNS
Bar O,Gelb S,Atamny K,Anzi S,Ben-Zvi A
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunocytochemistry
Wed Apr 01 00:00:00 EDT 2020
Stellate Cells, Hepatocytes, and Endothelial Cells Imprint the Kupffer Cell Identity on Monocytes Colonizing the Liver Macrophage Niche.
Immunity
Bonnardel J,T'Jonck W,Gaublomme D,Browaeys R,Scott CL,Martens L,Vanneste B,De Prijck S,Nedospasov SA,Kremer A,Van Hamme E,Borghgraef P,Toussaint W,De Bleser P,Mannaerts I,Beschin A,van Grunsven LA,Lambrecht BN,Taghon T,Lippens S,Elewaut D,Saeys Y,Guilliams M
13-1402-82 was used in Immunohistochemistry to show that the Kupffer cell niche is composed of stellate cells, hepatocytes, and endothelial cells that together imprint the liver-specific macrophage identity.
Tue Oct 15 00:00:00 EDT 2019
Standard screening methods underreport AAV-mediated transduction and gene editing.
Nature communications
Lang JF,Toulmin SA,Brida KL,Eisenlohr LC,Davidson BL
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunohistochemistry
Tue Jul 30 00:00:00 EDT 2019
Claudin-12 is not required for blood-brain barrier tight junction function.
Fluids and barriers of the CNS
Castro Dias M,Coisne C,Baden P,Enzmann G,Garrett L,Becker L,Hölter SM,Hrabě de Angelis M,Deutsch U,Engelhardt B
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Thu Sep 12 00:00:00 EDT 2019
Claudin-12 is not required for blood-brain barrier tight junction function.
Fluids and barriers of the CNS
Castro Dias M,Coisne C,Baden P,Enzmann G,Garrett L,Becker L,Hölter SM,Hrabě de Angelis M,Deutsch U,Engelhardt B
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Thu Sep 12 00:00:00 EDT 2019
Dual lineage tracing identifies intermediate mesenchymal stage for endocardial contribution to fibroblasts, coronary mural cells, and adipocytes.
The Journal of biological chemistry
Huang X,Feng T,Jiang Z,Meng J,Kou S,Lu Z,Chen W,Lin CP,Zhou B,Zhang H
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Fri May 31 00:00:00 EDT 2019
Dual lineage tracing identifies intermediate mesenchymal stage for endocardial contribution to fibroblasts, coronary mural cells, and adipocytes.
The Journal of biological chemistry
Huang X,Feng T,Jiang Z,Meng J,Kou S,Lu Z,Chen W,Lin CP,Zhou B,Zhang H
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Fri May 31 00:00:00 EDT 2019
Growth Factor Screening in Dystrophic Muscles Reveals PDGFB/PDGFRB-Mediated Migration of Interstitial Stem Cells.
International journal of molecular sciences
Camps J,Grosemans H,Gijsbers R,Maes C,Sampaolesi M
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Tue Mar 05 00:00:00 EST 2019
Label-Retaining, Putative Mesenchymal Stem Cells Contribute to Murine Myometrial Repair During Uterine Involution.
Stem cells and development
Patterson AL,George JW,Chatterjee A,Carpenter T,Wolfrum E,Pru JK,Teixeira JM
13-1402 was used in Immunohistochemistry-immunofluorescence to identify and characterize putative mesenchymal stem/progenitor cells in the murine myometrium using a combination of label retention and mesenchymal stem cell (MSC) marker expression and a pregnancy and uterine INV model.
Sat Dec 15 00:00:00 EST 2018
Molecular mechanisms of IL-33-mediated stromal interactions in cancer metastasis.
JCI insight
Andersson P,Yang Y,Hosaka K,Zhang Y,Fischer C,Braun H,Liu S,Yu G,Liu S,Beyaert R,Chang M,Li Q,Cao Y
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunohistochemistry
Thu Oct 18 00:00:00 EDT 2018
Pulmonary pericytes regulate lung morphogenesis.
Nature communications
Kato K,Diéguez-Hurtado R,Park DY,Hong SP,Kato-Azuma S,Adams S,Stehling M,Trappmann B,Wrana JL,Koh GY,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Fri Jun 22 00:00:00 EDT 2018
Repression of Interstitial Identity in Nephron Progenitor Cells by Pax2 Establishes the Nephron-Interstitium Boundary during Kidney Development.
Developmental cell
Naiman N,Fujioka K,Fujino M,Valerius MT,Potter SS,McMahon AP,Kobayashi A
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunohistochemistry (Frozen)
Mon May 22 00:00:00 EDT 2017
Repression of Interstitial Identity in Nephron Progenitor Cells by Pax2 Establishes the Nephron-Interstitium Boundary during Kidney Development.
Developmental cell
Naiman N,Fujioka K,Fujino M,Valerius MT,Potter SS,McMahon AP,Kobayashi A
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunohistochemistry (Frozen)
Mon May 22 00:00:00 EDT 2017
Smooth muscle cell recruitment to lymphatic vessels requires PDGFB and impacts vessel size but not identity.
Development (Cambridge, England)
Wang Y,Jin Y,Mäe MA,Zhang Y,Ortsäter H,Betsholtz C,Mäkinen T,Jakobsson L
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Sun Oct 01 00:00:00 EDT 2017
Laminin differentially regulates the stemness of type I and type II pericytes.
Stem cell research & therapy
Gautam J,Nirwane A,Yao Y
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Tue Feb 07 00:00:00 EST 2017
Laminin differentially regulates the stemness of type I and type II pericytes.
Stem cell research & therapy
Gautam J,Nirwane A,Yao Y
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Tue Feb 07 00:00:00 EST 2017
Pericytes regulate VEGF-induced endothelial sprouting through VEGFR1.
Nature communications
Eilken HM,Diéguez-Hurtado R,Schmidt I,Nakayama M,Jeong HW,Arf H,Adams S,Ferrara N,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunohistochemistry
Fri Nov 17 00:00:00 EST 2017
Neonatal pancreatic pericytes support β-cell proliferation.
Molecular metabolism
Epshtein A,Rachi E,Sakhneny L,Mizrachi S,Baer D,Landsman L
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Sun Oct 01 00:00:00 EDT 2017
Gas6 derived from cancer-associated fibroblasts promotes migration of Axl-expressing lung cancer cells during chemotherapy.
Scientific reports
Kanzaki R,Naito H,Kise K,Takara K,Eino D,Minami M,Shintani Y,Funaki S,Kawamura T,Kimura T,Okumura M,Takakura N
13-1402 was used in Flow cytometry/Cell sorting to suggest that the ligand Gas6 derived from cancer-associated fibroblasts is associated with poor clinical outcome, by promoting migration of Axl-expressing lung cancer cells during chemotherapy.
Wed Sep 06 00:00:00 EDT 2017
Age-dependent modulation of vascular niches for haematopoietic stem cells.
Nature
Kusumbe AP,Ramasamy SK,Itkin T,Mäe MA,Langen UH,Betsholtz C,Lapidot T,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Thu Apr 21 00:00:00 EDT 2016
Age-dependent modulation of vascular niches for haematopoietic stem cells.
Nature
Kusumbe AP,Ramasamy SK,Itkin T,Mäe MA,Langen UH,Betsholtz C,Lapidot T,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Thu Apr 21 00:00:00 EDT 2016
Age-dependent modulation of vascular niches for haematopoietic stem cells.
Nature
Kusumbe AP,Ramasamy SK,Itkin T,Mäe MA,Langen UH,Betsholtz C,Lapidot T,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Thu Apr 21 00:00:00 EDT 2016
Age-dependent modulation of vascular niches for haematopoietic stem cells.
Nature
Kusumbe AP,Ramasamy SK,Itkin T,Mäe MA,Langen UH,Betsholtz C,Lapidot T,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Thu Apr 21 00:00:00 EDT 2016
Paracrine Wnt1 Drives Interstitial Fibrosis without Inflammation by Tubulointerstitial Cross-Talk.
Journal of the American Society of Nephrology : JASN
Maarouf OH,Aravamudhan A,Rangarajan D,Kusaba T,Zhang V,Welborn J,Gauvin D,Hou X,Kramann R,Humphreys BD
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Tue Mar 01 00:00:00 EST 2016
Paracrine Wnt1 Drives Interstitial Fibrosis without Inflammation by Tubulointerstitial Cross-Talk.
Journal of the American Society of Nephrology : JASN
Maarouf OH,Aravamudhan A,Rangarajan D,Kusaba T,Zhang V,Welborn J,Gauvin D,Hou X,Kramann R,Humphreys BD
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Tue Mar 01 00:00:00 EST 2016
Epithelial cell integrin β1 is required for developmental angiogenesis in the pituitary gland.
Proceedings of the National Academy of Sciences of the United States of America
Scully KM,Skowronska-Krawczyk D,Krawczyk M,Merkurjev D,Taylor H,Livolsi A,Tollkuhn J,Stan RV,Rosenfeld MG
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunohistochemistry
Tue Nov 22 00:00:00 EST 2016
The role of pericytic laminin in blood brain barrier integrity maintenance.
Scientific reports
Gautam J,Zhang X,Yao Y
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Thu Nov 03 00:00:00 EDT 2016
The role of pericytic laminin in blood brain barrier integrity maintenance.
Scientific reports
Gautam J,Zhang X,Yao Y
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Thu Nov 03 00:00:00 EDT 2016
Laminin regulates PDGFRβ(+) cell stemness and muscle development.
Nature communications
Yao Y,Norris EH,Mason CE,Strickland S
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Tue May 03 00:00:00 EDT 2016
Heterogeneous fibroblasts underlie age-dependent tertiary lymphoid tissues in the kidney.
JCI insight
Sato Y,Mii A,Hamazaki Y,Fujita H,Nakata H,Masuda K,Nishiyama S,Shibuya S,Haga H,Ogawa O,Shimizu A,Narumiya S,Kaisho T,Arita M,Yanagisawa M,Miyasaka M,Sharma K,Minato N,Kawamoto H,Yanagita M
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Thu Jul 21 00:00:00 EDT 2016
The PDGF-BB-SOX7 axis-modulated IL-33 in pericytes and stromal cells promotes metastasis through tumour-associated macrophages.
Nature communications
Yang Y,Andersson P,Hosaka K,Zhang Y,Cao R,Iwamoto H,Yang X,Nakamura M,Wang J,Zhuang R,Morikawa H,Xue Y,Braun H,Beyaert R,Samani N,Nakae S,Hams E,Dissing S,Fallon PG,Langer R,Cao Y
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Fri May 06 00:00:00 EDT 2016
The PDGF-BB-SOX7 axis-modulated IL-33 in pericytes and stromal cells promotes metastasis through tumour-associated macrophages.
Nature communications
Yang Y,Andersson P,Hosaka K,Zhang Y,Cao R,Iwamoto H,Yang X,Nakamura M,Wang J,Zhuang R,Morikawa H,Xue Y,Braun H,Beyaert R,Samani N,Nakae S,Hams E,Dissing S,Fallon PG,Langer R,Cao Y
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Fri May 06 00:00:00 EDT 2016
Isoform-Specific Modulation of Inflammation Induced by Adenoviral Mediated Delivery of Platelet-Derived Growth Factors in the Adult Mouse Heart.
PloS one
Gallini R,Huusko J,Ylä-Herttuala S,Betsholtz C,Andrae J
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunohistochemistry
Mon Aug 07 00:00:00 EDT 2017
Analysis of the brain mural cell transcriptome.
Scientific reports
He L,Vanlandewijck M,Raschperger E,Andaloussi Mäe M,Jung B,Lebouvier T,Ando K,Hofmann J,Keller A,Betsholtz C
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Tue Oct 11 00:00:00 EDT 2016
Analysis of the brain mural cell transcriptome.
Scientific reports
He L,Vanlandewijck M,Raschperger E,Andaloussi Mäe M,Jung B,Lebouvier T,Ando K,Hofmann J,Keller A,Betsholtz C
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Tue Oct 11 00:00:00 EDT 2016
Endothelial cells are progenitors of cardiac pericytes and vascular smooth muscle cells.
Nature communications
Chen Q,Zhang H,Liu Y,Adams S,Eilken H,Stehling M,Corada M,Dejana E,Zhou B,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Fri Aug 12 00:00:00 EDT 2016
Endothelial cells are progenitors of cardiac pericytes and vascular smooth muscle cells.
Nature communications
Chen Q,Zhang H,Liu Y,Adams S,Eilken H,Stehling M,Corada M,Dejana E,Zhou B,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Fri Aug 12 00:00:00 EDT 2016
Mesodermal iPSC-derived progenitor cells functionally regenerate cardiac and skeletal muscle.
The Journal of clinical investigation
Quattrocelli M,Swinnen M,Giacomazzi G,Camps J,Barthélemy I,Ceccarelli G,Caluwé E,Grosemans H,Thorrez L,Pelizzo G,Muijtjens M,Verfaillie CM,Blot S,Janssens S,Sampaolesi M
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2015
The αvβ1 integrin plays a critical in vivo role in tissue fibrosis.
Science translational medicine
Reed NI,Jo H,Chen C,Tsujino K,Arnold TD,DeGrado WF,Sheppard D
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Wed May 20 00:00:00 EDT 2015
Regional and stage-specific effects of prospectively purified vascular cells on the adult V-SVZ neural stem cell lineage.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Crouch EE,Liu C,Silva-Vargas V,Doetsch F
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Wed Mar 18 00:00:00 EDT 2015
Regional and stage-specific effects of prospectively purified vascular cells on the adult V-SVZ neural stem cell lineage.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Crouch EE,Liu C,Silva-Vargas V,Doetsch F
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Wed Mar 18 00:00:00 EDT 2015
Regional and stage-specific effects of prospectively purified vascular cells on the adult V-SVZ neural stem cell lineage.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Crouch EE,Liu C,Silva-Vargas V,Doetsch F
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Wed Mar 18 00:00:00 EDT 2015
Fibroblast Growth Factor 9 Imparts Hierarchy and Vasoreactivity to the Microcirculation of Renal Tumors and Suppresses Metastases.
The Journal of biological chemistry
Yin H,Frontini MJ,Arpino JM,Nong Z,O'Neil C,Xu Y,Balint B,Ward AD,Chakrabarti S,Ellis CG,Gros R,Pickering JG
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Neutralization
Fri Sep 04 00:00:00 EDT 2015
Fibroblast Growth Factor 9 Imparts Hierarchy and Vasoreactivity to the Microcirculation of Renal Tumors and Suppresses Metastases.
The Journal of biological chemistry
Yin H,Frontini MJ,Arpino JM,Nong Z,O'Neil C,Xu Y,Balint B,Ward AD,Chakrabarti S,Ellis CG,Gros R,Pickering JG
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Neutralization
Fri Sep 04 00:00:00 EDT 2015
Fibroblast Growth Factor 9 Imparts Hierarchy and Vasoreactivity to the Microcirculation of Renal Tumors and Suppresses Metastases.
The Journal of biological chemistry
Yin H,Frontini MJ,Arpino JM,Nong Z,O'Neil C,Xu Y,Balint B,Ward AD,Chakrabarti S,Ellis CG,Gros R,Pickering JG
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Neutralization
Fri Sep 04 00:00:00 EDT 2015
Pericytes are progenitors for coronary artery smooth muscle.
eLife
Volz KS,Jacobs AH,Chen HI,Poduri A,McKay AS,Riordan DP,Kofler N,Kitajewski J,Weissman I,Red-Horse K
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunohistochemistry
Mon Oct 19 00:00:00 EDT 2015
Mfsd2a is critical for the formation and function of the blood-brain barrier.
Nature
Ben-Zvi A,Lacoste B,Kur E,Andreone BJ,Mayshar Y,Yan H,Gu C
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Thu May 22 00:00:00 EDT 2014
Osterix marks distinct waves of primitive and definitive stromal progenitors during bone marrow development.
Developmental cell
Mizoguchi T,Pinho S,Ahmed J,Kunisaki Y,Hanoun M,Mendelson A,Ono N,Kronenberg HM,Frenette PS
13-1402 was used in Flow cytometry/Cell sorting to investigate the developmental origin of stromal cells, showing that osterix marks distinct waves of stromal progenitors during bone marrow development.
Mon May 12 00:00:00 EDT 2014
Formation of persistent hyperplastic primary vitreous in ephrin-A5-/- mice.
Investigative ophthalmology & visual science
Son AI,Sheleg M,Cooper MA,Sun Y,Kleiman NJ,Zhou R
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Wed Mar 19 00:00:00 EDT 2014
Distribution of microsomal prostaglandin E synthase-1 in the mouse brain.
The Journal of comparative neurology
Eskilsson A,Tachikawa M,Hosoya K,Blomqvist A
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Wed Oct 01 00:00:00 EDT 2014
The number of fetal nephron progenitor cells limits ureteric branching and adult nephron endowment.
Cell reports
Cebrian C,Asai N,D'Agati V,Costantini F
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunohistochemistry
Thu Apr 10 00:00:00 EDT 2014
Astrocytic laminin regulates pericyte differentiation and maintains blood brain barrier integrity.
Nature communications
Yao Y,Chen ZL,Norris EH,Strickland S
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Mon Mar 03 00:00:00 EST 2014
Wnt4/β-catenin signaling in medullary kidney myofibroblasts.
Journal of the American Society of Nephrology : JASN
DiRocco DP,Kobayashi A,Taketo MM,McMahon AP,Humphreys BD
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Sun Sep 01 00:00:00 EDT 2013
Ephrin-B2 controls PDGFRβ internalization and signaling.
Genes & development
Nakayama A,Nakayama M,Turner CJ,Höing S,Lepore JJ,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Sun Dec 01 00:00:00 EST 2013
Ephrin-B2 controls PDGFRβ internalization and signaling.
Genes & development
Nakayama A,Nakayama M,Turner CJ,Höing S,Lepore JJ,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Sun Dec 01 00:00:00 EST 2013
Ephrin-B2 controls PDGFRβ internalization and signaling.
Genes & development
Nakayama A,Nakayama M,Turner CJ,Höing S,Lepore JJ,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Sun Dec 01 00:00:00 EST 2013
High glucose and diabetes modulate cellular proteasome function: Implications in the pathogenesis of diabetes complications.
Biochemical and biophysical research communications
Aghdam SY,Gurel Z,Ghaffarieh A,Sorenson CM,Sheibani N
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Fri Mar 08 00:00:00 EST 2013
Pericytes derived from adipose-derived stem cells protect against retinal vasculopathy.
PloS one
Mendel TA,Clabough EB,Kao DS,Demidova-Rice TN,Durham JT,Zotter BC,Seaman SA,Cronk SM,Rakoczy EP,Katz AJ,Herman IM,Peirce SM,Yates PA
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Fri Jan 10 00:00:00 EST 2014
The bHLH transcription factor Tcf21 is required for lineage-specific EMT of cardiac fibroblast progenitors.
Development (Cambridge, England)
Acharya A,Baek ST,Huang G,Eskiocak B,Goetsch S,Sung CY,Banfi S,Sauer MF,Olsen GS,Duffield JS,Olson EN,Tallquist MD
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Fri Jun 01 00:00:00 EDT 2012
Multiple stromal populations contribute to pulmonary fibrosis without evidence for epithelial to mesenchymal transition.
Proceedings of the National Academy of Sciences of the United States of America
Rock JR,Barkauskas CE,Cronce MJ,Xue Y,Harris JR,Liang J,Noble PW,Hogan BL
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Tue Dec 27 00:00:00 EST 2011
Differential contribution to neuroendocrine tumorigenesis of parallel egfr signaling in cancer cells and pericytes.
Genes & cancer
Nolan-Stevaux O,Truitt MC,Pahler JC,Olson P,Guinto C,Lee DC,Hanahan D
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2010
Six2 defines and regulates a multipotent self-renewing nephron progenitor population throughout mammalian kidney development.
Cell stem cell
Kobayashi A,Valerius MT,Mugford JW,Carroll TJ,Self M,Oliver G,McMahon AP
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 13-1402-82) in Immunofluorescence
Thu Aug 07 00:00:00 EDT 2008