Product References
PDGFRβ+ cells play a dual role as hematopoietic precursors and niche cells during mouse ontogeny.
Cell reports
Sá da Bandeira D,Kilpatrick AM,Marques M,Gomez-Salazar M,Ventura T,Gonzalez ZN,Stefancova D,Rossi F,Vermeren M,Vink CS,Beltran M,Henderson NC,Jung B,van der Linden R,van de Werken HJG,van Ijcken WFJ,Betsholtz C,Forbes SJ,Cuervo H,Crisan M
17-1402-82 was used in Flow cytometry/Cell sorting to demonstrate that PDGFRβ+ cells play a dual role in murine hematopoiesis.
Tue Jul 19 00:00:00 EDT 2022
Heterogeneous bone-marrow stromal progenitors drive myelofibrosis via a druggable alarmin axis.
Cell stem cell
Leimkühler NB,Gleitz HFE,Ronghui L,Snoeren IAM,Fuchs SNR,Nagai JS,Banjanin B,Lam KH,Vogl T,Kuppe C,Stalmann USA,Büsche G,Kreipe H,Gütgemann I,Krebs P,Banz Y,Boor P,Tai EW,Brümmendorf TH,Koschmieder S,Crysandt M,Bindels E,Kramann R,Costa IG,Schneider RK
17-1402-82 was used in Flow Cytometry to develop a comprehensive map of the stroma in myeloproliferative neoplasms (MPNs)on a single-cell level in murine models and patient samples.
Thu Apr 01 00:00:00 EDT 2021
Cellular dormancy in minimal residual disease following targeted therapy.
Breast cancer research : BCR
Ruth JR,Pant DK,Pan TC,Seidel HE,Baksh SC,Keister BA,Singh R,Sterner CJ,Bakewell SJ,Moody SE,Belka GK,Chodosh LA
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Fri Jun 04 00:00:00 EDT 2021
Cellular dormancy in minimal residual disease following targeted therapy.
Breast cancer research : BCR
Ruth JR,Pant DK,Pan TC,Seidel HE,Baksh SC,Keister BA,Singh R,Sterner CJ,Bakewell SJ,Moody SE,Belka GK,Chodosh LA
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Fri Jun 04 00:00:00 EDT 2021
Distinct Fibroblast Lineages Give Rise to NG2+ Pericyte Populations in Mouse Skin Development and Repair.
Frontiers in cell and developmental biology
Goss G,Rognoni E,Salameti V,Watt FM
17-1402-82 was used in Flow Cytometry to investigate if Ng2+ perivascular cells represent a heterogeneous lineage restricted population and if it is primarily recruited from the papillary or reticular fibroblast lineages during tissue regeneration.
Wed Mar 02 00:00:00 EST 2022
Distinct Fibroblast Lineages Give Rise to NG2+ Pericyte Populations in Mouse Skin Development and Repair.
Frontiers in cell and developmental biology
Goss G,Rognoni E,Salameti V,Watt FM
17-1402-82 was used in Flow Cytometry to investigate if Ng2+ perivascular cells represent a heterogeneous lineage restricted population and if it is primarily recruited from the papillary or reticular fibroblast lineages during tissue regeneration.
Wed Mar 02 00:00:00 EST 2022
Direct contribution of skeletal muscle mesenchymal progenitors to bone repair.
Nature communications
Julien A,Kanagalingam A,Martínez-Sarrà E,Megret J,Luka M,Ménager M,Relaix F,Colnot C
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Immunohistochemistry
Mon May 17 00:00:00 EDT 2021
Direct contribution of skeletal muscle mesenchymal progenitors to bone repair.
Nature communications
Julien A,Kanagalingam A,Martínez-Sarrà E,Megret J,Luka M,Ménager M,Relaix F,Colnot C
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Immunohistochemistry
Mon May 17 00:00:00 EDT 2021
Cardiac pericytes function as key vasoactive cells to regulate homeostasis and disease.
FEBS open bio
Lee LL,Khakoo AY,Chintalgattu V
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
Heterogeneity of murine periosteum progenitors involved in fracture healing.
eLife
Matthews BG,Novak S,Sbrana FV,Funnell JL,Cao Y,Buckels EJ,Grcevic D,Kalajzic I
17-1402-82 was used in Flow Cytometry to demonstrate that the periosteum is highly enriched with skeletal progenitor cells, and there is heterogeneity in the populations of cells that contribute to mature lineages during periosteal fracture healing.
Tue Feb 09 00:00:00 EST 2021
Metastasis-initiating cells induce and exploit a fibroblast niche to fuel malignant colonization of the lungs.
Nature communications
Pein M,Insua-Rodríguez J,Hongu T,Riedel A,Meier J,Wiedmann L,Decker K,Essers MAG,Sinn HP,Spaich S,Sütterlin M,Schneeweiss A,Trumpp A,Oskarsson T
17-1402-82 was used in Flow Cytometry to show that disseminated breast cancer cells evoke phenotypic changes in lung fibroblasts, forming a supportive metastatic niche.
Fri Mar 20 00:00:00 EDT 2020
Dual lineage tracing identifies intermediate mesenchymal stage for endocardial contribution to fibroblasts, coronary mural cells, and adipocytes.
The Journal of biological chemistry
Huang X,Feng T,Jiang Z,Meng J,Kou S,Lu Z,Chen W,Lin CP,Zhou B,Zhang H
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Fri May 31 00:00:00 EDT 2019
Growth Factor Screening in Dystrophic Muscles Reveals PDGFB/PDGFRB-Mediated Migration of Interstitial Stem Cells.
International journal of molecular sciences
Camps J,Grosemans H,Gijsbers R,Maes C,Sampaolesi M
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Tue Mar 05 00:00:00 EST 2019
Bone marrow drives central nervous system regeneration after radiation injury.
The Journal of clinical investigation
Dietrich J,Baryawno N,Nayyar N,Valtis YK,Yang B,Ly I,Besnard A,Severe N,Gustafsson KU,Andronesi OC,Batchelor TT,Sahay A,Scadden DT
17-1402 was used in Flow cytometry/Cell sorting to demonstrate that the hematopoietic system provides critical regenerative drive to the brain that can be modulated by clinically available agents.
Tue Jan 02 00:00:00 EST 2018
Pulmonary pericytes regulate lung morphogenesis.
Nature communications
Kato K,Diéguez-Hurtado R,Park DY,Hong SP,Kato-Azuma S,Adams S,Stehling M,Trappmann B,Wrana JL,Koh GY,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Fri Jun 22 00:00:00 EDT 2018
Pulmonary pericytes regulate lung morphogenesis.
Nature communications
Kato K,Diéguez-Hurtado R,Park DY,Hong SP,Kato-Azuma S,Adams S,Stehling M,Trappmann B,Wrana JL,Koh GY,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Fri Jun 22 00:00:00 EDT 2018
Gpr124 is essential for blood-brain barrier integrity in central nervous system disease.
Nature medicine
Chang J,Mancuso MR,Maier C,Liang X,Yuki K,Yang L,Kwong JW,Wang J,Rao V,Vallon M,Kosinski C,Zhang JJ,Mah AT,Xu L,Li L,Gholamin S,Reyes TF,Li R,Kuhnert F,Han X,Yuan J,Chiou SH,Brettman AD,Daly L,Corney DC,Cheshier SH,Shortliffe LD,Wu X,Snyder M,Chan P,Giffard RG,Chang HY,Andreasson K,Kuo CJ
17-1402 was used in Flow cytometry/Cell sorting to examine the function of the endothelial G-protein-coupled receptor Gpr124 in homeostasis and pathology in adult mice.
Sat Apr 01 00:00:00 EDT 2017
Differential cytokine contributions of perivascular haematopoietic stem cell niches.
Nature cell biology
Asada N,Kunisaki Y,Pierce H,Wang Z,Fernandez NF,Birbrair A,Ma'ayan A,Frenette PS
17-1402 was used in Flow cytometry/Cell sorting to demonstrate that the effects of cytokines regulating haematopoietic stem cell functions are dependent on the producing cell sources.
Wed Mar 01 00:00:00 EST 2017
Cardiac Fibroblasts Adopt Osteogenic Fates and Can Be Targeted to Attenuate Pathological Heart Calcification.
Cell stem cell
Pillai ICL,Li S,Romay M,Lam L,Lu Y,Huang J,Dillard N,Zemanova M,Rubbi L,Wang Y,Lee J,Xia M,Liang O,Xie YH,Pellegrini M,Lusis AJ,Deb A
17-1402 was used in Flow cytometry/Cell sorting to study the cell identify and mechanisms contributing to pathological heart muscle calcification.
Thu Feb 02 00:00:00 EST 2017
Laminin differentially regulates the stemness of type I and type II pericytes.
Stem cell research & therapy
Gautam J,Nirwane A,Yao Y
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Tue Feb 07 00:00:00 EST 2017
Neonatal pancreatic pericytes support β-cell proliferation.
Molecular metabolism
Epshtein A,Rachi E,Sakhneny L,Mizrachi S,Baer D,Landsman L
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Sun Oct 01 00:00:00 EDT 2017
Pericytes regulate VEGF-induced endothelial sprouting through VEGFR1.
Nature communications
Eilken HM,Diéguez-Hurtado R,Schmidt I,Nakayama M,Jeong HW,Arf H,Adams S,Ferrara N,Adams RH
17-1402 was used in Flow cytometry/Cell sorting to show that pericytes promote endothelial sprouting in the postnatal retinal vasculature.
Fri Nov 17 00:00:00 EST 2017
Age-dependent modulation of vascular niches for haematopoietic stem cells.
Nature
Kusumbe AP,Ramasamy SK,Itkin T,Mäe MA,Langen UH,Betsholtz C,Lapidot T,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Thu Apr 21 00:00:00 EDT 2016
Paracrine Wnt1 Drives Interstitial Fibrosis without Inflammation by Tubulointerstitial Cross-Talk.
Journal of the American Society of Nephrology : JASN
Maarouf OH,Aravamudhan A,Rangarajan D,Kusaba T,Zhang V,Welborn J,Gauvin D,Hou X,Kramann R,Humphreys BD
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2016
Analysis of the brain mural cell transcriptome.
Scientific reports
He L,Vanlandewijck M,Raschperger E,Andaloussi Mäe M,Jung B,Lebouvier T,Ando K,Hofmann J,Keller A,Betsholtz C
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Tue Oct 11 00:00:00 EDT 2016
Endothelial cells are progenitors of cardiac pericytes and vascular smooth muscle cells.
Nature communications
Chen Q,Zhang H,Liu Y,Adams S,Eilken H,Stehling M,Corada M,Dejana E,Zhou B,Adams RH
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Fri Aug 12 00:00:00 EDT 2016
Mesodermal iPSC-derived progenitor cells functionally regenerate cardiac and skeletal muscle.
The Journal of clinical investigation
Quattrocelli M,Swinnen M,Giacomazzi G,Camps J,Barthélemy I,Ceccarelli G,Caluwé E,Grosemans H,Thorrez L,Pelizzo G,Muijtjens M,Verfaillie CM,Blot S,Janssens S,Sampaolesi M
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2015
Regional and stage-specific effects of prospectively purified vascular cells on the adult V-SVZ neural stem cell lineage.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Crouch EE,Liu C,Silva-Vargas V,Doetsch F
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Wed Mar 18 00:00:00 EDT 2015
Pericytes derived from adipose-derived stem cells protect against retinal vasculopathy.
PloS one
Mendel TA,Clabough EB,Kao DS,Demidova-Rice TN,Durham JT,Zotter BC,Seaman SA,Cronk SM,Rakoczy EP,Katz AJ,Herman IM,Peirce SM,Yates PA
17-1402 was used in Flow cytometry/Cell sorting to examine the ability of ASCs to differentiate into pericytes that can stabilize retinal vessels in retinal vasculopathy models.
Fri Jan 10 00:00:00 EST 2014
Differential contribution to neuroendocrine tumorigenesis of parallel egfr signaling in cancer cells and pericytes.
Genes & cancer
Nolan-Stevaux O,Truitt MC,Pahler JC,Olson P,Guinto C,Lee DC,Hanahan D
Published figure using CD140b (PDGFRB) monoclonal antibody (Product # 17-1402-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2010