Product References
Development and characterization of a new monoclonal antibody against SARS-CoV-2 NSP12 (RdRp).
Journal of medical virology
Meng W,Guo S,Cao S,Shuda M,Robinson-McCarthy LR,McCarthy KR,Shuda Y,Paniz Mondolfi AE,Bryce C,Grimes Z,Sordillo EM,Cordon-Cardo C,Li P,Zhang H,Perlman S,Guo H,Gao SJ,Chang Y,Moore PS
14-1449-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot to suggest that NSP12 may have diminished steady-state expression or extensive posttranslation modifications that limit antibody reactivity during SARS-CoV-2 replication.
Sun Jan 01 00:00:00 UTC 2023
Development and characterization of a new monoclonal antibody against SARS-CoV-2 NSP12 (RdRp).
Journal of medical virology
Meng W,Guo S,Cao S,Shuda M,Robinson-McCarthy LR,McCarthy KR,Shuda Y,Paniz Mondolfi AE,Bryce C,Grimes Z,Sordillo EM,Cordon-Cardo C,Li P,Zhang H,Perlman S,Guo H,Gao SJ,Chang Y,Moore PS
14-1449-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot to suggest that NSP12 may have diminished steady-state expression or extensive posttranslation modifications that limit antibody reactivity during SARS-CoV-2 replication.
Sun Jan 01 00:00:00 UTC 2023
Endothelial-to-mesenchymal transition: a precursor to pulmonary arterial remodelling in patients with idiopathic pulmonary fibrosis.
ERJ open research
Gaikwad AV,Lu W,Dey S,Bhattarai P,Haug G,Larby J,Chia C,Jaffar J,Westall G,Singhera GK,Hackett TL,Eapen MS,Sohal SS
14-1449-82 was used in Immunohistochemistry to provide evidence for active endothelial-to-mesenchymal transition (EndMT) in idiopathic pulmonary fibrosis patients.
Wed Mar 01 00:00:00 UTC 2023
Vascular remodelling in idiopathic pulmonary fibrosis patients and its detrimental effect on lung physiology: potential role of endothelial-to-mesenchymal transition.
ERJ open research
Gaikwad AV,Lu W,Dey S,Bhattarai P,Chia C,Larby J,Haug G,Myers S,Jaffar J,Westall G,Singhera GK,Hackett TL,Markos J,Eapen MS,Sohal SS
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Immunohistochemistry (Paraffin)
Sat Jan 01 00:00:00 UTC 2022
Long-term cultured microvascular networks on chip for tumor vascularization research and drug testing.
Biomicrofluidics
Zhang K,Du Z,Yuan T,Huang J,Zhao X,Mi S
14-1449-82 was used in Immunocytochemistry-immunoflourescence to construct a mimicking breast cancer invasion model based on a microfluidic chip platform, and the maintenance time of the self-assembled microvascular networks significantly improved by culturing with fibroblasts (up to 13 days).
Fri Jul 01 00:00:00 UTC 2022
In Situ Fabrication and Perfusion of Tissue-Engineered Blood Vessel Microphysiological System.
Methods in molecular biology (Clifton, N.J.)
Zhang X,Abutaleb NO,Salmon E,Truskey GA
14-1449-82 was used in Immunocytochemistry-immunofluorescence to develop a novel device to fabricate arteriole-scale human endothelialized TEBVs in situ with smaller volumes and higher throughput than previously reported.
Wed Jan 05 00:00:00 UTC 2022
Lymphatic-specific intracellular modulation of receptor tyrosine kinase signaling improves lymphatic growth and function.
Science signaling
Kataru RP,Baik JE,Park HJ,Ly CL,Shin J,Schwartz N,Lu TT,Ortega S,Mehrara BJ
14-1449-82 was used in Western Blot to indicate that intracellular modulation of lymphangiogenesis is effective in inducing functional lymphatic networks and has no off-target inflammatory effects.
Tue Aug 10 00:00:00 UTC 2021
Upregulation of ETV2 Expression Promotes Endothelial Differentiation of Human Dental Pulp Stem Cells.
Cell transplantation
Li J,Zhu Y,Li N,Wu T,Zheng X,Heng BC,Zou D,Xu J
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Tue Dec 21 00:00:00 UTC 2021
Deciphering the Role of Extracellular Vesicles Derived from ZIKV-Infected hcMEC/D3 Cells on the Blood-Brain Barrier System.
Viruses
Fikatas A,Dehairs J,Noppen S,Doijen J,Vanderhoydonc F,Meyen E,Swinnen JV,Pannecouque C,Schols D
14-1449-82 was used in Immunocytochemistry-immunoflourescence to investigate the role of extracellular vesicles derived from ZIKV-infected human brain microvascular endothelial cells on the blood-brain barrier (BBB) system.
Thu Nov 25 00:00:00 UTC 2021
Endothelial specific YY1 deletion restricts tumor angiogenesis and tumor growth.
Scientific reports
Liu H,Qiu Y,Pei X,Chitteti R,Steiner R,Zhang S,Jin ZG
14-1449-82 was used in Immunohistochemistry-immunofluorescence to demonstrate that endothelial YY1 has a crucial role in tumor angiogenesis and suggest that targeting endothelial YY1 could be a potential therapeutic strategy for cancer treatment.
Tue Nov 24 00:00:00 UTC 2020
Endothelial specific YY1 deletion restricts tumor angiogenesis and tumor growth.
Scientific reports
Liu H,Qiu Y,Pei X,Chitteti R,Steiner R,Zhang S,Jin ZG
14-1449-82 was used in Immunohistochemistry-immunofluorescence to demonstrate that endothelial YY1 has a crucial role in tumor angiogenesis and suggest that targeting endothelial YY1 could be a potential therapeutic strategy for cancer treatment.
Tue Nov 24 00:00:00 UTC 2020
Targeting QKI-7 in vivo restores endothelial cell function in diabetes.
Nature communications
Yang C,Eleftheriadou M,Kelaini S,Morrison T,González MV,Caines R,Edwards N,Yacoub A,Edgar K,Moez A,Ivetic A,Zampetaki A,Zeng L,Wilkinson FL,Lois N,Stitt AW,Grieve DJ,Margariti A
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Thu Jul 30 00:00:00 UTC 2020
Altered endothelial ETB receptor expression in postmenopausal women.
American journal of physiology. Heart and circulatory physiology
Kuczmarski AV,Shoemaker LN,Hobson JC,Edwards DG,Wenner MM
14-1449-82 was used in Immunohistochemistry-immunofluorescence to provide additional mechanistic insight into vascular endothelial dysfunction in postmenopausal women.
Wed Jul 01 00:00:00 UTC 2020
Human Cardiac Organoids for Modeling Genetic Cardiomyopathy.
Cells
Filippo Buono M,von Boehmer L,Strang J,Hoerstrup SP,Emmert MY,Nugraha B
14-1449-82 was used in Immunocytochemistry to demonstrate the generation of cardiac organoids using a triculture approach of human induced pluripotent stem-cell-derived cardiomyocytes (hiPS-CMs)- from healthy subjects and cardiomyopathy patients-human cardiac microvascular endothelial cells (HCMECs) and human cardiac fibroblasts (HCFs).
Mon Jul 20 00:00:00 UTC 2020
Vascular Tumor Recapitulated in Endothelial Cells from hiPSCs Engineered to Express the SERPINE1-FOSB Translocation.
Cell reports. Medicine
van IJzendoorn DGP,Salvatori DCF,Cao X,van den Hil F,Briaire-de Bruijn IH,de Jong D,Mei H,Mummery CL,Szuhai K,Bovée JVMG,Orlova VV
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Tue Dec 22 00:00:00 UTC 2020
Isolating pulmonary microvascular endothelial cells ex vivo: Implications for pulmonary arterial hypertension, and a caution on the use of commercial biomaterials.
PloS one
Wertheim BM,Lin YD,Zhang YY,Samokhin AO,Alba GA,Arons E,Yu PB,Maron BA
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Mon Nov 04 00:00:00 UTC 2019
Angiogenic and Restorative Abilities of Human Mesenchymal Stem Cells Were Reduced Following Treatment With Serum From Diabetes Mellitus Type 2 Patients.
Journal of cellular biochemistry
Rezaie J,Mehranjani MS,Rahbarghazi R,Shariatzadeh MA
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Mon Jan 01 00:00:00 UTC 2018
Enhanced Therapeutic and Long-Term Dynamic Vascularization Effects of Human Pluripotent Stem Cell-Derived Endothelial Cells Encapsulated in a Nanomatrix Gel.
Circulation
Lee SJ,Sohn YD,Andukuri A,Kim S,Byun J,Han JW,Park IH,Jun HW,Yoon YS
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Tue Nov 14 00:00:00 UTC 2017
Generating high-purity cardiac and endothelial derivatives from patterned mesoderm using human pluripotent stem cells.
Nature protocols
Palpant NJ,Pabon L,Friedman CE,Roberts M,Hadland B,Zaunbrecher RJ,Bernstein I,Zheng Y,Murry CE
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Sun Jan 01 00:00:00 UTC 2017
MMP-2 and MMP-14 Silencing Inhibits VEGFR2 Cleavage and Induces the Differentiation of Porcine Adipose-Derived Mesenchymal Stem Cells to Endothelial Cells.
Stem cells translational medicine
Almalki SG,Llamas Valle Y,Agrawal DK
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Mon May 01 00:00:00 UTC 2017
ERK signaling is required for VEGF-A/VEGFR2-induced differentiation of porcine adipose-derived mesenchymal stem cells into endothelial cells.
Stem cell research & therapy
Almalki SG,Agrawal DK
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Fri May 12 00:00:00 UTC 2017
Challenges in enumeration of CTCs in breast cancer using techniques independent of cytokeratin expression.
PloS one
Castle J,Morris K,Pritchard S,Kirwan CC
Thu Apr 27 00:00:00 UTC 2017
Challenges in enumeration of CTCs in breast cancer using techniques independent of cytokeratin expression.
PloS one
Castle J,Morris K,Pritchard S,Kirwan CC
Thu Apr 27 00:00:00 UTC 2017
Challenges in enumeration of CTCs in breast cancer using techniques independent of cytokeratin expression.
PloS one
Castle J,Morris K,Pritchard S,Kirwan CC
Thu Apr 27 00:00:00 UTC 2017
Challenges in enumeration of CTCs in breast cancer using techniques independent of cytokeratin expression.
PloS one
Castle J,Morris K,Pritchard S,Kirwan CC
Thu Apr 27 00:00:00 UTC 2017
Targeted Disruption of TCF12 Reveals HEB as Essential in Human Mesodermal Specification and Hematopoiesis.
Stem cell reports
Li Y,Brauer PM,Singh J,Xhiku S,Yoganathan K,Zúñiga-Pflücker JC,Anderson MK
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Tue Sep 12 00:00:00 UTC 2017
Targeted Disruption of TCF12 Reveals HEB as Essential in Human Mesodermal Specification and Hematopoiesis.
Stem cell reports
Li Y,Brauer PM,Singh J,Xhiku S,Yoganathan K,Zúñiga-Pflücker JC,Anderson MK
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Tue Sep 12 00:00:00 UTC 2017
Tumourigenic non-small-cell lung cancer mesenchymal circulating tumour cells: a clinical case study.
Annals of oncology : official journal of the European Society for Medical Oncology
Morrow CJ,Trapani F,Metcalf RL,Bertolini G,Hodgkinson CL,Khandelwal G,Kelly P,Galvin M,Carter L,Simpson KL,Williamson S,Wirth C,Simms N,Frankliln L,Frese KK,Rothwell DG,Nonaka D,Miller CJ,Brady G,Blackhall FH,Dive C
Wed Jun 01 00:00:00 UTC 2016
Tumourigenic non-small-cell lung cancer mesenchymal circulating tumour cells: a clinical case study.
Annals of oncology : official journal of the European Society for Medical Oncology
Morrow CJ,Trapani F,Metcalf RL,Bertolini G,Hodgkinson CL,Khandelwal G,Kelly P,Galvin M,Carter L,Simpson KL,Williamson S,Wirth C,Simms N,Frankliln L,Frese KK,Rothwell DG,Nonaka D,Miller CJ,Brady G,Blackhall FH,Dive C
Wed Jun 01 00:00:00 UTC 2016
Rab11a Mediates Vascular Endothelial-Cadherin Recycling and Controls Endothelial Barrier Function.
Arteriosclerosis, thrombosis, and vascular biology
Yan Z,Wang ZG,Segev N,Hu S,Minshall RD,Dull RO,Zhang M,Malik AB,Hu G
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Immunocytochemistry
Mon Feb 01 00:00:00 UTC 2016
Rab11a Mediates Vascular Endothelial-Cadherin Recycling and Controls Endothelial Barrier Function.
Arteriosclerosis, thrombosis, and vascular biology
Yan Z,Wang ZG,Segev N,Hu S,Minshall RD,Dull RO,Zhang M,Malik AB,Hu G
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Immunocytochemistry
Mon Feb 01 00:00:00 UTC 2016
Rab11a Mediates Vascular Endothelial-Cadherin Recycling and Controls Endothelial Barrier Function.
Arteriosclerosis, thrombosis, and vascular biology
Yan Z,Wang ZG,Segev N,Hu S,Minshall RD,Dull RO,Zhang M,Malik AB,Hu G
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Immunocytochemistry
Mon Feb 01 00:00:00 UTC 2016
Definitive Hematopoietic Multipotent Progenitor Cells Are Transiently Generated From Hemogenic Endothelial Cells in Human Pluripotent Stem Cells.
Journal of cellular physiology
Bai H,Liu Y,Xie Y,Hoyle DL,Brodsky RA,Cheng L,Cheng T,Wang ZZ
14144982 was used in flow cytometry to define a differentiation system to generate definitive hematopoietic multipotent progenitor cells from hemato-endothelial progenitors via the endothelial monolayer
Sun May 01 00:00:00 UTC 2016
Mitochondria-Targeted Antioxidant SkQR1 Reduces TNF-Induced Endothelial Permeability in vitro.
Biochemistry. Biokhimiia
Galkin II,Pletjushkina OY,Zinovkin RA,Zakharova VV,Chernyak BV,Popova EN
14144982 was used in immunocytochemistry and western blot to study the role of TNF in endothelial cell permeability and apoptosis.
Sat Oct 01 00:00:00 UTC 2016
Vasculogenic mimicry in small cell lung cancer.
Nature communications
Williamson SC,Metcalf RL,Trapani F,Mohan S,Antonello J,Abbott B,Leong HS,Chester CP,Simms N,Polanski R,Nonaka D,Priest L,Fusi A,Carlsson F,Carlsson A,Hendrix MJ,Seftor RE,Seftor EA,Rothwell DG,Hughes A,Hicks J,Miller C,Kuhn P,Brady G,Simpson KL,Blackhall FH,Dive C
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Immunohistochemistry
Wed Nov 09 00:00:00 UTC 2016
Vasculogenic mimicry in small cell lung cancer.
Nature communications
Williamson SC,Metcalf RL,Trapani F,Mohan S,Antonello J,Abbott B,Leong HS,Chester CP,Simms N,Polanski R,Nonaka D,Priest L,Fusi A,Carlsson F,Carlsson A,Hendrix MJ,Seftor RE,Seftor EA,Rothwell DG,Hughes A,Hicks J,Miller C,Kuhn P,Brady G,Simpson KL,Blackhall FH,Dive C
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Immunohistochemistry
Wed Nov 09 00:00:00 UTC 2016
Vasculogenic mimicry in small cell lung cancer.
Nature communications
Williamson SC,Metcalf RL,Trapani F,Mohan S,Antonello J,Abbott B,Leong HS,Chester CP,Simms N,Polanski R,Nonaka D,Priest L,Fusi A,Carlsson F,Carlsson A,Hendrix MJ,Seftor RE,Seftor EA,Rothwell DG,Hughes A,Hicks J,Miller C,Kuhn P,Brady G,Simpson KL,Blackhall FH,Dive C
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Immunohistochemistry
Wed Nov 09 00:00:00 UTC 2016
Vasculogenic mimicry in small cell lung cancer.
Nature communications
Williamson SC,Metcalf RL,Trapani F,Mohan S,Antonello J,Abbott B,Leong HS,Chester CP,Simms N,Polanski R,Nonaka D,Priest L,Fusi A,Carlsson F,Carlsson A,Hendrix MJ,Seftor RE,Seftor EA,Rothwell DG,Hughes A,Hicks J,Miller C,Kuhn P,Brady G,Simpson KL,Blackhall FH,Dive C
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Immunohistochemistry
Wed Nov 09 00:00:00 UTC 2016
Direct conversion of human amniotic cells into endothelial cells without transitioning through a pluripotent state.
Nature protocols
Ginsberg M,Schachterle W,Shido K,Rafii S
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Tue Dec 01 00:00:00 UTC 2015
Direct conversion of human amniotic cells into endothelial cells without transitioning through a pluripotent state.
Nature protocols
Ginsberg M,Schachterle W,Shido K,Rafii S
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Tue Dec 01 00:00:00 UTC 2015
Mesenchymal stromal cells form vascular tubes when placed in fibrin sealant and accelerate wound healing in vivo.
Biomaterials
Mendez JJ,Ghaedi M,Sivarapatna A,Dimitrievska S,Shao Z,Osuji CO,Steinbacher DM,Leffell DJ,Niklason LE
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Sun Feb 01 00:00:00 UTC 2015
Fc gamma receptor IIb participates in maternal IgG trafficking of human placental endothelial cells.
International journal of molecular medicine
Ishikawa T,Takizawa T,Iwaki J,Mishima T,Ui-Tei K,Takeshita T,Matsubara S,Takizawa T
14-1449 was used in Immunocytochemistry to suggest that FCGR2B2 compartments participate in the transcytosis of maternal IgG across the human placental endothelium.
Fri May 01 00:00:00 UTC 2015
Histone Demethylases KDM4A and KDM4C Regulate Differentiation of Embryonic Stem Cells to Endothelial Cells.
Stem cell reports
Wu L,Wary KK,Revskoy S,Gao X,Tsang K,Komarova YA,Rehman J,Malik AB
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Tue Jul 14 00:00:00 UTC 2015
Induced Pluripotent Stem Cells to Model Human Fibrodysplasia Ossificans Progressiva.
Stem cell reports
Cai J,Orlova VV,Cai X,Eekhoff EMW,Zhang K,Pei D,Pan G,Mummery CL,Ten Dijke P
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Tue Dec 08 00:00:00 UTC 2015
PDGFRα and CD51 mark human nestin+ sphere-forming mesenchymal stem cells capable of hematopoietic progenitor cell expansion.
The Journal of experimental medicine
Pinho S,Lacombe J,Hanoun M,Mizoguchi T,Bruns I,Kunisaki Y,Frenette PS
14-1449 was used in Immunofluorescence to evaluate putative cell surface mesenchymal stem cell markers to identify Nestin+ cells in the mouse and human bone marrow.
Mon Jul 01 00:00:00 UTC 2013
MicroRNA-16 affects key functions of human endothelial progenitor cells.
Journal of leukocyte biology
Goretti E,Rolland-Turner M,Léonard F,Zhang L,Wagner DR,Devaux Y
14-1449 was used in Flow cytometry/Cell sorting to determine the role of miRNAs in human endothelial progenitor cells.
Wed May 01 00:00:00 UTC 2013
Efficient direct reprogramming of mature amniotic cells into endothelial cells by ETS factors and TGFβ suppression.
Cell
Ginsberg M,James D,Ding BS,Nolan D,Geng F,Butler JM,Schachterle W,Pulijaal VR,Mathew S,Chasen ST,Xiang J,Rosenwaks Z,Shido K,Elemento O,Rabbany SY,Rafii S
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Fri Oct 26 00:00:00 UTC 2012
Identification of the hemogenic endothelial progenitor and its direct precursor in human pluripotent stem cell differentiation cultures.
Cell reports
Choi KD,Vodyanik MA,Togarrati PP,Suknuntha K,Kumar A,Samarjeet F,Probasco MD,Tian S,Stewart R,Thomson JA,Slukvin II
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1449-82) in Flow Cytometry
Thu Sep 27 00:00:00 UTC 2012
Neovascular niche for human myeloma cells in immunodeficient mouse bone.
PloS one
Iriuchishima H,Takubo K,Miyakawa Y,Nakamura-Ishizu A,Miyauchi Y,Fujita N,Miyamoto K,Miyamoto T,Ikeda E,Kizaki M,Nojima Y,Suda T
14-1449 was used in Immunocytochemistry to provide a useful system to analyze the dynamics of MM pathophysiology, interactions of MM cells with other cellular compartments, and the utility of novel anti-MM therapies.
Tue Jul 31 00:00:00 UTC 2012
Neovascular niche for human myeloma cells in immunodeficient mouse bone.
PloS one
Iriuchishima H,Takubo K,Miyakawa Y,Nakamura-Ishizu A,Miyauchi Y,Fujita N,Miyamoto K,Miyamoto T,Ikeda E,Kizaki M,Nojima Y,Suda T
14-1449 was used in Immunocytochemistry to provide a useful system to analyze the dynamics of MM pathophysiology, interactions of MM cells with other cellular compartments, and the utility of novel anti-MM therapies.
Tue Jul 31 00:00:00 UTC 2012
Identification of endothelial cell junctional proteins and lymphocyte receptors involved in transendothelial migration of human effector memory CD4+ T cells.
Journal of immunology (Baltimore, Md. : 1950)
Manes TD,Pober JS
14-1449 was used in Blocking experiments to further analyze effector memory (EM) CD4(+) T cell migrational responses to either chemokine or TCR-activating signals.
Tue Feb 01 00:00:00 UTC 2011
High harvest yield, high expansion, and phenotype stability of CD146 mesenchymal stromal cells from whole primitive human umbilical cord tissue.
Journal of biomedicine & biotechnology
Schugar RC,Chirieleison SM,Wescoe KE,Schmidt BT,Askew Y,Nance JJ,Evron JM,Peault B,Deasy BM
14-1449 was used in Flow cytometry/Cell sorting to show novel in situ characterization to identify and localize a panel of some markers expressed by mesenchymal stromal cells and CD146 in the umbilical cord.
Wed Mar 10 00:00:00 UTC 2010
Differential requirements for hematopoietic commitment between human and rhesus embryonic stem cells.
Stem cells (Dayton, Ohio)
Rajesh D,Chinnasamy N,Mitalipov SM,Wolf DP,Slukvin I,Thomson JA,Shaaban AF
Thu Feb 01 00:00:00 UTC 2007