Product References
The matrix microenvironment influences but does not dominate tissue-specific stem cell lineage differentiation.
Materials today. Bio
Pei YA,Mikaeiliagah E,Wang B,Zhang X,Pei M
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Fri Dec 01 00:00:00 UTC 2023
Reprogramming bone progenitor identity and potency through control of collagen density and oxygen tension.
iScience
Al Hosni R,Bozec L,Roberts SJ,Cheema U
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Fri Apr 15 00:00:00 UTC 2022
LGR5 expressing skin fibroblasts define a major cellular hub perturbed in scleroderma.
Cell
Gur C,Wang SY,Sheban F,Zada M,Li B,Kharouf F,Peleg H,Aamar S,Yalin A,Kirschenbaum D,Braun-Moscovici Y,Jaitin DA,Meir-Salame T,Hagai E,Kragesteen BK,Avni B,Grisariu S,Bornstein C,Shlomi-Loubaton S,David E,Shreberk-Hassidim R,Molho-Pessach V,Amar D,Tzur T,Kuint R,Gross M,Barboy O,Moshe A,Fellus-Alyagor L,Hirsch D,Addadi Y,Erenfeld S,Biton M,Tzemach T,Elazary A,Naparstek Y,Tzemach R,Weiner A,Giladi A,Balbir-Gurman A,Amit I
12-1469-42 was used in Flow cytometry/Cell sorting to find immune compartment dysfunction only in a specific subtype of diffuse Systemic sclerosis (scleroderma, SSc) patients but global dysregulation of the stromal compartment, particularly in a previously undefined subset of LGR5+-scleroderma-associated fibroblasts (ScAFs).
Thu Apr 14 00:00:00 UTC 2022
Matrix reverses immortalization-mediated stem cell fate determination.
Biomaterials
Wang Y,Hu G,Hill RC,Dzieciatkowska M,Hansen KC,Zhang XB,Yan Z,Pei M
12-1469-42 was used in Flow Cytometry to provide a promising model for in-depth investigation of the matrix protein influence on surrounding stem cell differentiation.
Fri Jan 01 00:00:00 UTC 2021
uPAR+ extracellular vesicles: a robust biomarker of resistance to checkpoint inhibitor immunotherapy in metastatic melanoma patients.
Journal for immunotherapy of cancer
Porcelli L,Guida M,De Summa S,Di Fonte R,De Risi I,Garofoli M,Caputo M,Negri A,Strippoli S,Serratì S,Azzariti A
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Sat May 01 00:00:00 UTC 2021
uPAR+ extracellular vesicles: a robust biomarker of resistance to checkpoint inhibitor immunotherapy in metastatic melanoma patients.
Journal for immunotherapy of cancer
Porcelli L,Guida M,De Summa S,Di Fonte R,De Risi I,Garofoli M,Caputo M,Negri A,Strippoli S,Serratì S,Azzariti A
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Sat May 01 00:00:00 UTC 2021
Chitinase 3-like-1 contributes to acetaminophen-induced liver injury by promoting hepatic platelet recruitment.
eLife
Shan Z,Li L,Atkins CL,Wang M,Wen Y,Jeong J,Moreno NF,Feng D,Gui X,Zhang N,Lee CG,Elias JA,Lee WM,Gao B,Lam FW,An Z,Ju C
12-1469-42 was used in Flow Cytometry to unveil a critical role of Chi3l1 in hepatic platelet recruitment during AILI.
Thu Jun 10 00:00:00 UTC 2021
Site-Dependent Lineage Preference of Adipose Stem Cells.
Frontiers in cell and developmental biology
Wang T,Hill RC,Dzieciatkowska M,Zhu L,Infante AM,Hu G,Hansen KC,Pei M
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Wed Dec 14 00:00:00 UTC 2022
Site-Dependent Lineage Preference of Adipose Stem Cells.
Frontiers in cell and developmental biology
Wang T,Hill RC,Dzieciatkowska M,Zhu L,Infante AM,Hu G,Hansen KC,Pei M
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Wed Dec 14 00:00:00 UTC 2022
Aging affects responsiveness of peripheral blood mononuclear cells to immunosuppression of periodontal ligament stem cells.
The Journal of international medical research
Li X,Zhang B,Wang H,Zhao X,Zhang Z,Ding G,Wei F
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2020
Aging affects responsiveness of peripheral blood mononuclear cells to immunosuppression of periodontal ligament stem cells.
The Journal of international medical research
Li X,Zhang B,Wang H,Zhao X,Zhang Z,Ding G,Wei F
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2020
Apoptotic bodies derived from mesenchymal stem cells promote cutaneous wound healing via regulating the functions of macrophages.
Stem cell research & therapy
Liu J,Qiu X,Lv Y,Zheng C,Dong Y,Dou G,Zhu B,Liu A,Wang W,Zhou J,Liu S,Liu S,Gao B,Jin Y
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Fri Nov 27 00:00:00 UTC 2020
Apoptotic bodies derived from mesenchymal stem cells promote cutaneous wound healing via regulating the functions of macrophages.
Stem cell research & therapy
Liu J,Qiu X,Lv Y,Zheng C,Dong Y,Dou G,Zhu B,Liu A,Wang W,Zhou J,Liu S,Liu S,Gao B,Jin Y
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Fri Nov 27 00:00:00 UTC 2020
Intrinsic transcriptomic sex differences in human endothelial cells at birth and in adults are associated with coronary artery disease targets.
Scientific reports
Hartman RJG,Kapteijn DMC,Haitjema S,Bekker MN,Mokry M,Pasterkamp G,Civelek M,den Ruijter HM
12-1469 was used in Flow cytometry/Cell sorting to identify the transcriptomic sex differences in endothelial cells at birth and in adults.
Thu Jul 23 00:00:00 UTC 2020
Impact of Fibronectin Knockout on Proliferation and Differentiation of Human Infrapatellar Fat Pad-Derived Stem Cells.
Frontiers in bioengineering and biotechnology
Wang Y,Fu Y,Yan Z,Zhang XB,Pei M
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Wed Dec 14 00:00:00 UTC 2022
LPS‑induced upregulation of the TLR4 signaling pathway inhibits osteogenic differentiation of human periodontal ligament stem cells under inflammatory conditions.
International journal of molecular medicine
Yu B,Li Q,Zhou M
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Sat Jun 01 00:00:00 UTC 2019
Combination of immunogenic oncolytic adenovirus ONCOS-102 with anti-PD-1 pembrolizumab exhibits synergistic antitumor effect in humanized A2058 melanoma huNOG mouse model.
Oncoimmunology
Kuryk L,Møller AW,Jaderberg M
12-1469 was used in Flow cytometry/Cell sorting to investigate the cytotoxicity of ONCOS-102 and pembrolizumab, an anti-PD-1 antibody, in four human melanoma cell lines, A375, A2058, SK-Mel-2 and SK-Mel-28.
Tue Jan 12 00:00:00 UTC 2021
Molecular Programming of Perivascular Stem Cell Precursors.
Stem cells (Dayton, Ohio)
Yianni V,Sharpe PT
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Sat Dec 01 00:00:00 UTC 2018
Extracellular vesicles from human saliva promote hemostasis by delivering coagulant tissue factor to activated platelets.
Journal of thrombosis and haemostasis : JTH
Yu Y,Gool E,Berckmans RJ,Coumans FAW,Barendrecht AD,Maas C,van der Wel NN,Altevogt P,Sturk A,Nieuwland R
12-1469 was used in Flow cytometry/Cell sorting to demonstrate that extracellular vesicles from human saliva facilitate homeostasis at sites of skin injury through exposure of coagulants TF and CD24.
Fri Jun 01 00:00:00 UTC 2018
Alkaline Phosphatase Controls Lineage Switching of Mesenchymal Stem Cells by Regulating the LRP6/GSK3β Complex in Hypophosphatasia.
Theranostics
Liu W,Zhang L,Xuan K,Hu C,Li L,Zhang Y,Jin F,Jin Y
12-1469 was used in Flow cytometry/Cell sorting to elucidate a novel role of tissue-nonspecific alkaline phosphatase in controlling bone marrow mesenchymal stem cell lineage selection.
Mon Sep 09 00:00:00 UTC 2019
Tumor-associated B-cells induce tumor heterogeneity and therapy resistance.
Nature communications
Somasundaram R,Zhang G,Fukunaga-Kalabis M,Perego M,Krepler C,Xu X,Wagner C,Hristova D,Zhang J,Tian T,Wei Z,Liu Q,Garg K,Griss J,Hards R,Maurer M,Hafner C,Mayerhöfer M,Karanikas G,Jalili A,Bauer-Pohl V,Weihsengruber F,Rappersberger K,Koller J,Lang R,Hudgens C,Chen G,Tetzlaff M,Wu L,Frederick DT,Scolyer RA,Long GV,Damle M,Ellingsworth C,Grinman L,Choi H,Gavin BJ,Dunagin M,Raj A,Scholler N,Gross L,Beqiri M,Bennett K,Watson I,Schaider H,Davies MA,Wargo J,Czerniecki BJ,Schuchter L,Herlyn D,Flaherty K,Herlyn M,Wagner SN
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Tue Sep 19 00:00:00 UTC 2017
CD44-mediated monocyte transmigration across Cryptococcus neoformans-infected brain microvascular endothelial cells is enhanced by HIV-1 gp41-I90 ectodomain.
Journal of biomedical science
He X,Shi X,Puthiyakunnon S,Zhang L,Zeng Q,Li Y,Boddu S,Qiu J,Lai Z,Ma C,Xie Y,Long M,Du L,Huang SH,Cao H
12-1469 was used in Immunopurification to examine the future prospects to inflammatory responses induced by HIV-1 gp41-I90 ectodomain and to effectively eliminate the opportunistic infections in AIDS patients.
Sat Feb 20 00:00:00 UTC 2016
Analysis of Bone-Cartilage-Stromal Progenitor Populations in Trauma Induced and Genetic Models of Heterotopic Ossification.
Stem cells (Dayton, Ohio)
Agarwal S,Loder SJ,Sorkin M,Li S,Shrestha S,Zhao B,Mishina Y,James AW,Levi B
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Wed Jun 01 00:00:00 UTC 2016
Generation of embryonic stem cells from mouse adipose-tissue derived cells via somatic cell nuclear transfer.
Cell cycle (Georgetown, Tex.)
Qin Y,Qin J,Zhou C,Li J,Gao WQ
12-1469 was used in Flow cytometry/Cell sorting to demonstrates that ESCs can be generated from the adipose tissue by somatic cell nuclear transfer (SCNT) and suggests that ADCs can be a new donor-cell type for potential therapeutic cloning.
Tue Jan 26 00:00:00 UTC 2016
Repositioning of Memantine as a Potential Novel Therapeutic Agent against Meningitic E. coli-Induced Pathogenicities through Disease-Associated Alpha7 Cholinergic Pathway and RNA Sequencing-Based Transcriptome Analysis of Host Inflammatory Responses.
PloS one
Yu JY,Zhang B,Peng L,Wu CH,Cao H,Zhong JF,Hoffman J,Huang SH
12-1469-42 was used in Flow Cytometry to suggest that memantine can efficiently block host inflammatory responses to bacterial infection through modulation of both inflammatory and anti-inflammatory pathways.
Mon Apr 11 00:00:00 UTC 2016
Kupffer cells potentiate liver sinusoidal endothelial cell injury in sepsis by ligating programmed cell death ligand-1.
Journal of leukocyte biology
Hutchins NA,Wang F,Wang Y,Chung CS,Ayala A
12-1469 was used in Flow cytometry/Cell sorting to study Kupffer cell-mediated potentiation of liver sinusoidal endothelial cell injury in sepsis, by ligating programmed cell death ligand-1.
Fri Nov 01 00:00:00 UTC 2013
Application of anodized titanium for enhanced recruitment of endothelial progenitor cells.
Nanoscale research letters
Moradi I,Behjati M,Kazemi M
Published figure using CD146 monoclonal antibody (Product # 12-1469-42) in Flow Cytometry
Thu Jun 07 00:00:00 UTC 2012