Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: The monoclonal antibody HP-MA4 recognizes several CD158 proteins, also known as KIR2D, specifically KIR2DL1 (CD158a), KIR2DS1 (CD158h), KIR2DS3, and KIR2DS5 (CD158g). It does not recognize KIR2DL2 (CD158b1), KIR2DS2 (CD158j), KIR2DL4 (CD158d) or KIR2DS4 (CD158i) and crossreactivity to KIR2DL3 (CD158b2) and KIR2DL5 (CD158f) are unknown. CD158 proteins are members of the killer cell immunoglobulin-like receptors (KIR) family of transmembrane glycoproteins that are expressed on NK cells and a subset of T cells. Each KIR has an isoform with either a short (S) or long (L) cytoplasmic domain which transduces either an activating or inhibitory signal, respectively. The crossreactivity of the HP-MA4 antibody to KIR2DS3 and KIR2DS5 was published in 2013.
Applications Reported: This HP-MA4 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This HP-MA4 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.
Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-822-49) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-54) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several "framework" genes are found in all haplotypes. The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals. The ligands for several KIR proteins are subsets of HLA class I molecules; thus, KIR proteins are thought to play an important role in regulation of the immune response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: CD158; CD158 antigen-like family member A; CD158 antigen-like family member D; CD158 antigen-like family member G; CD158 antigen-like family member H; CD158 antigen-like family member I; CD158 antigen-like family member J; CD158a; CD158B1; CD158d; CD158g; CD158h; CD158i; CD158j; G9P; Killer cell immunoglobulin-like receptor 2DL1; Killer cell immunoglobulin-like receptor 2DL4; Killer cell immunoglobulin-like receptor 2DS1; Killer cell immunoglobulin-like receptor 2DS2; Killer cell immunoglobulin-like receptor 2DS4; Killer cell immunoglobulin-like receptor 2DS5; killer cell immunoglobulin-like receptor KIR2DS2; killer cell immunoglobulin-like receptor KIRDS1; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 4; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5; Killer cell inhibitory receptor 103AS; killer Ig receptor; killer inhibitory receptor 2-2-1; killer inhibitory receptor 4-1-2; killer-cell Ig-like receptor; killer-cell immunoglobulin-like receptor; killer-cell immunoglobulin-like receptor 2DL4; killer-cell immunoglobulin-like receptor two domains short tail 2 protein; KIR antigen 2DS4; KIR-103AS; MHC class I NK cell receptor; MHC class I NK cell receptor Eb6 ActI; MHC class I NK cell receptor KIR103AS; natural killer associated transcript 5; natural killer cell inhibitory receptor; natural killer cell receptor; Natural killer-associated transcript 1; Natural killer-associated transcript 5; Natural killer-associated transcript 8; Natural killer-associated transcript 9; NK cell receptor; NK receptor 183 ActI; NKAT-1; NKAT-5; NKAT-8; NKAT-9; NKAT6; p50 killer cell activating receptor KAR-K1d; p50.3; p58 killer cell inhibitory receptor KIR-K64; p58 killer cell inhibitory receptor KIR-K7a; p58 natural killer cell receptor clone CL-49; P58 natural killer cell receptor clones CL-39/CL-17; p58 natural killer cell receptor clones CL-42/47.11; p58 NK cell inhibitory receptor NKR-K6; p58 NK receptor CL-39/CL-17; p58 NK receptor CL-42/47.11; p58 NK receptor CL-49; p58.1 MHC class-I-specific NK receptor; XXbac-BCX195L8.1
Gene Aliases: 183ActI; CD158A; CD158b; CD158D; CD158G; CD158H; CD158I; CD158J; cl-49; G9P; KIR; KIR-103AS; KIR-2DL4; KIR-2DS2; KIR-2DS4; KIR-K64; KIR103; KIR103AS; KIR1D; KIR221; KIR2DL1; KIR2DL4; KIR2DP1DL1; KIR2DS1; KIR2DS2; KIR2DS4; KIR2DS5; KIR412; KKA3; NKAT; NKAT-1; NKAT-5; NKAT-8; NKAT1; NKAT5; NKAT8; NKAT9; p50.1; p58.1
UniProt ID: (Human) P43626, (Human) Q99706, (Human) Q14954, (Human) P43631, (Human) P43632, (Human) Q14953
Entrez Gene ID: (Human) 3802, (Human) 3805, (Human) 3806, (Human) 100132285, (Human) 3809, (Human) 3810
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support