Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This antibody has specificity for Mouse CD16-2/FCGR4.
FcgRIV, also known as CD16-2, is an intermediate-affinity activating receptor for IgG2a and IgG2b. CD16-2 is the mouse homolog of human FcgRIIIA. CD16-2 is a low-affinity IgE receptor for all allotypes and the ligation of FcgRIV by antigen-IgE immune complexes promotes macrophage-mediated phagocytosis and is involved in lung inflammation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD16-2; CD16a; Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity III, receptor for (CD16); Fc gammaRIV; Fc receptor-like 3; FcgammaRIV; IgG Fc receptor III-A; Low affinity immunoglobulin gamma Fc region receptor III-A
Gene Aliases: 4833442P21Rik; CD16-2; FcgammaRIV; Fcgr3a; Fcgr4; FcgRIV; Fcrl3
UniProt ID: (Mouse) A0A0B4J1G0
Entrez Gene ID: (Mouse) 246256
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support