Product References
Designing Synthetic, Sulfated Glycosaminoglycan Mimetics That Are Orally Bioavailable and Exhibiting In Vivo Anticancer Activity.
Journal of medicinal chemistry
Morla S,Ravikumar O,O'Hara C,Boothello R,Vera A,Abdelfadiel EI,Fayyad R,Afosah DK,Sharon C,Fernandez L,Shah SA,Patel BB,Desai UR
Published figure using CD184 (CXCR4) monoclonal antibody (Product # 12-9991-82) in Flow Cytometry
Thu Jan 26 00:00:00 UTC 2023
Age-induced alterations of granulopoiesis generate atypical neutrophils that aggravate stroke pathology.
Nature immunology
Gullotta GS,De Feo D,Friebel E,Semerano A,Scotti GM,Bergamaschi A,Butti E,Brambilla E,Genchi A,Capotondo A,Gallizioli M,Coviello S,Piccoli M,Vigo T,Della Valle P,Ronchi P,Comi G,D'Angelo A,Maugeri N,Roveri L,Uccelli A,Becher B,Martino G,Bacigaluppi M
12-9991-82 was used in Flow Cytometry to investigate the impact of age-related changes in the immune system on stroke.
Thu Jun 01 00:00:00 UTC 2023
PHGDH is required for germinal center formation and is a therapeutic target in MYC-driven lymphoma.
The Journal of clinical investigation
D'Avola A,Legrave N,Tajan M,Chakravarty P,Shearer RL,King HW,Kluckova K,Cheung EC,Clear AJ,Gunawan AS,Zhang L,James LK,MacRae JI,Gribben JG,Calado DP,Vousden KH,Riches JC
Published figure using CD184 (CXCR4) monoclonal antibody (Product # 12-9991-82) in Flow Cytometry
Mon May 02 00:00:00 UTC 2022
Mesenchymal stromal cells equipped by IFNα empower T cells with potent anti-tumor immunity.
Oncogene
Zhang T,Wang Y,Li Q,Lin L,Xu C,Xue Y,Hu M,Shi Y,Wang Y
12-9991-82 was used in Flow Cytometry to conclude that IFNα-MSCs represent a novel tumor immunotherapy strategy, especially when combined with PD-L1 blockade.
Tue Mar 01 00:00:00 UTC 2022
Krüppel-like factor 2 controls IgA plasma cell compartmentalization and IgA responses.
Mucosal immunology
Wittner J,Schulz SR,Steinmetz TD,Berges J,Hauke M,Channell WM,Cunningham AF,Hauser AE,Hutloff A,Mielenz D,Jäck HM,Schuh W
Published figure using CD184 (CXCR4) monoclonal antibody (Product # 12-9991-82) in Flow Cytometry
Fri Apr 01 00:00:00 UTC 2022
SETD2 Haploinsufficiency Enhances Germinal Center-Associated AICDA Somatic Hypermutation to Drive B-cell Lymphomagenesis.
Cancer discovery
Leung W,Teater M,Durmaz C,Meydan C,Chivu AG,Chadburn A,Rice EJ,Muley A,Camarillo JM,Arivalagan J,Li Z,Flowers CR,Kelleher NL,Danko CG,Imielinski M,Dave SS,Armstrong SA,Mason CE,Melnick AM
12-9991-82 was used in Flow cytometry/Cell sorting to define a B cell-specific oncogenic effect of SETD2 heterozygous mutation, which unleashes AICDA mutagenesis of nontemplate strand DNA in the GC reaction, resulting in lymphomas with heavy mutational burden.
Wed Jul 06 00:00:00 UTC 2022
Neutrophil "plucking" on megakaryocytes drives platelet production and boosts cardiovascular disease.
Immunity
Petzold T,Zhang Z,Ballesteros I,Saleh I,Polzin A,Thienel M,Liu L,Ul Ain Q,Ehreiser V,Weber C,Kilani B,Mertsch P,Götschke J,Cremer S,Fu W,Lorenz M,Ishikawa-Ankerhold H,Raatz E,El-Nemr S,Görlach A,Marhuenda E,Stark K,Pircher J,Stegner D,Gieger C,Schmidt-Supprian M,Gaertner F,Almendros I,Kelm M,Schulz C,Hidalgo A,Massberg S
12-9991-82 was used in Flow Cytometry to establish neutrophil plucking as a target to reduce thromboischemic events.
Tue Dec 13 00:00:00 UTC 2022
Smc3 dosage regulates B cell transit through germinal centers and restricts their malignant transformation.
Nature immunology
Rivas MA,Meydan C,Chin CR,Challman MF,Kim D,Bhinder B,Kloetgen A,Viny AD,Teater MR,McNally DR,Doane AS,Béguelin W,Fernández MTC,Shen H,Wang X,Levine RL,Chen Z,Tsirigos A,Elemento O,Mason CE,Melnick AM
Published figure using CD184 (CXCR4) monoclonal antibody (Product # 12-9991-82) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2021
Cell-permeable transgelin-2 as a potent therapeutic for dendritic cell-based cancer immunotherapy.
Journal of hematology & oncology
Kim HR,Park JS,Park JH,Yasmin F,Kim CH,Oh SK,Chung IJ,Jun CD
12-9991 was used in Flow cytometry/Cell sorting to indicate that cell-permeable transgelin-2 have a potential clinical value as a cancer immunotherapy based on DCs.
Wed Mar 17 00:00:00 UTC 2021
Inhibition of Rag GTPase signaling in mice suppresses B cell responses and lymphomagenesis with minimal detrimental trade-offs.
Cell reports
Ortega-Molina A,Lebrero-Fernández C,Sanz A,Deleyto-Seldas N,Plata-Gómez AB,Menéndez C,Graña-Castro O,Caleiras E,Efeyan A
12-9991-82 was used in Flow Cytometry to support the efficacy and safety of moderate inhibition of nutrient signaling against pathological B cells.
Tue Jul 13 00:00:00 UTC 2021
WASp Is Crucial for the Unique Architecture of the Immunological Synapse in Germinal Center B-Cells.
Frontiers in cell and developmental biology
Li Y,Bhanja A,Upadhyaya A,Zhao X,Song W
12-9991-82 was used in Flow Cytometry to reveal essential roles for WASp in the generation and maintenance of unique structures for GCB immunological synapses.
Mon Feb 28 00:00:00 UTC 2022
TBL1XR1 Mutations Drive Extranodal Lymphoma by Inducing a Pro-tumorigenic Memory Fate.
Cell
Venturutti L,Teater M,Zhai A,Chadburn A,Babiker L,Kim D,Béguelin W,Lee TC,Kim Y,Chin CR,Yewdell WT,Raught B,Phillip JM,Jiang Y,Staudt LM,Green MR,Chaudhuri J,Elemento O,Farinha P,Weng AP,Nissen MD,Steidl C,Morin RD,Scott DW,Privé GG,Melnick AM
12-9991-82 was used in Flow Cytometry to show that TBL1XR1 mutations skew the humoral immune response toward generating abnormal immature memory B cells (MB), while impairing plasma cell differentiation.
Thu Jul 23 00:00:00 UTC 2020
SARS-CoV-2 mRNA Vaccines Foster Potent Antigen-Specific Germinal Center Responses Associated with Neutralizing Antibody Generation.
Immunity
Lederer K,Castaño D,Gómez Atria D,Oguin TH,Wang S,Manzoni TB,Muramatsu H,Hogan MJ,Amanat F,Cherubin P,Lundgreen KA,Tam YK,Fan SHY,Eisenlohr LC,Maillard I,Weissman D,Bates P,Krammer F,Sempowski GD,Pardi N,Locci M
Published figure using CD184 (CXCR4) monoclonal antibody (Product # 12-9991-82) in Flow Cytometry
Tue Dec 15 00:00:00 UTC 2020
Mutant EZH2 Induces a Pre-malignant Lymphoma Niche by Reprogramming the Immune Response.
Cancer cell
Béguelin W,Teater M,Meydan C,Hoehn KB,Phillip JM,Soshnev AA,Venturutti L,Rivas MA,Calvo-Fernández MT,Gutierrez J,Camarillo JM,Takata K,Tarte K,Kelleher NL,Steidl C,Mason CE,Elemento O,Allis CD,Kleinstein SH,Melnick AM
12-9991 was used in Flow cytometry/Cell sorting to show that EZH2 mutations initiate FL by attenuating germinal center(GC) B cell requirement for T cell help and driving slow expansion of GC centrocytes that become enmeshed with and dependent on follicular dendritic cells.
Mon May 11 00:00:00 UTC 2020
Inhibition of Migration, Invasion and Drug Resistance of Pancreatic Adenocarcinoma Cells - Role of Snail, Slug and Twist and Small Molecule Inhibitors.
OncoTargets and therapy
Kaşıkcı E,Aydemir E,Bayrak ÖF,Şahin F
Published figure using CD184 (CXCR4) monoclonal antibody (Product # 12-9991-82) in Flow Cytometry
Fri May 20 00:00:00 UTC 2022
B Cell Speed and B-FDC Contacts in Germinal Centers Determine Plasma Cell Output via Swiprosin-1/EFhd2.
Cell reports
Reimer D,Meyer-Hermann M,Rakhymzhan A,Steinmetz T,Tripal P,Thomas J,Boettcher M,Mougiakakos D,Schulz SR,Urbanczyk S,Hauser AE,Niesner RA,Mielenz D
12-9991 was used in Flow cytometry/Cell sorting to explain how germinal centre B cells integrate speed and persistence and directionality of cell migration.
Tue Aug 11 00:00:00 UTC 2020
Sleep modulates haematopoiesis and protects against atherosclerosis.
Nature
McAlpine CS,Kiss MG,Rattik S,He S,Vassalli A,Valet C,Anzai A,Chan CT,Mindur JE,Kahles F,Poller WC,Frodermann V,Fenn AM,Gregory AF,Halle L,Iwamoto Y,Hoyer FF,Binder CJ,Libby P,Tafti M,Scammell TE,Nahrendorf M,Swirski FK
12-9991-82 was used in Flow Cytometry to investigate the cellular and molecular mechanisms by which sleep maintains cardiovascular health.
Fri Feb 01 00:00:00 UTC 2019
The Bone Marrow Protects and Optimizes Immunological Memory during Dietary Restriction.
Cell
Collins N,Han SJ,Enamorado M,Link VM,Huang B,Moseman EA,Kishton RJ,Shannon JP,Dixit D,Schwab SR,Hickman HD,Restifo NP,McGavern DB,Schwartzberg PL,Belkaid Y
12-9991-82 was used in Flow Cytometry to show that memory T cells collapsed in secondary lymphoid organs in the context of dietary restriction (DR) but dramatically accumulated within the bone marrow (BM), where they adopted a state associated with energy conservation.
Thu Aug 22 00:00:00 UTC 2019
Enhanced germinal center reaction by targeting vaccine antigen to major histocompatibility complex class II molecules.
NPJ vaccines
Andersen TK,Huszthy PC,Gopalakrishnan RP,Jacobsen JT,Fauskanger M,Tveita AA,Grødeland G,Bogen B
Published figure using CD184 (CXCR4) monoclonal antibody (Product # 12-9991-82) in Flow Cytometry
Thu Oct 05 00:00:00 UTC 2023
Neutrophil Cytosolic Factor 1 in Dendritic Cells Promotes Autoreactive CD8+ T Cell Activation via Cross-Presentation in Type 1 Diabetes.
Frontiers in immunology
Liu C,Whitener RL,Lin A,Xu Y,Chen J,Savinov A,Leiding JW,Wallet MA,Mathews CE
12-9991-82 was used in Flow Cytometry to demonstrate that Ncf1 (p47phox) is required for activation and effector function of CD8+ T cells by acting both intrinsically within the T cell as well as within professional antigen presenting cells.
Wed Sep 09 00:00:00 UTC 2020
Nidogen-1 Contributes to the Interaction Network Involved in Pro-B Cell Retention in the Peri-sinusoidal Hematopoietic Stem Cell Niche.
Cell reports
Balzano M,De Grandis M,Vu Manh TP,Chasson L,Bardin F,Farina A,Sergé A,Bidaut G,Charbord P,Hérault L,Bailly AL,Cartier-Michaud A,Boned A,Dalod M,Duprez E,Genever P,Coles M,Bajenoff M,Xerri L,Aurrand-Lions M,Schiff C,Mancini SJC
12-9991 was used in Flow cytometry/Cell sorting to study pro-B cells in their support of stroll cells and their interaction network allowing their retention.
Tue Mar 19 00:00:00 UTC 2019
Cbl Ubiquitin Ligases Control B Cell Exit from the Germinal-Center Reaction.
Immunity
Li X,Gadzinsky A,Gong L,Tong H,Calderon V,Li Y,Kitamura D,Klein U,Langdon WY,Hou F,Zou YR,Gu H
12-9991-82 was used in Flow Cytometry to show that deleting E3 ubiquitin ligases Cbl and Cbl-b (Cbls) in GC B cells results in the early exit of high-affinity antigen-specific B cells from the GC reaction and thus impaired clonal expansion.
Tue Mar 20 00:00:00 UTC 2018
Immunological effects of a low-dose cytarabine, aclarubicin and granulocyte-colony stimulating factor priming regimen on a mouse leukemia model.
Oncology letters
Chen J,Yang N,Liu H,Yao H,Wang J,Yang Y,Zhang W
Sat Sep 01 00:00:00 UTC 2018
B Cell Receptor Crosslinking Augments Germinal Center B Cell Selection when T Cell Help Is Limiting.
Cell reports
Turner JS,Ke F,Grigorova IL
12-9991-82 was used in Flow cytometry/Cell sorting to investigate how T cells influence germinal center B cell expansion and plasmablast formation.
Tue Nov 06 00:00:00 UTC 2018
A Functional Bioluminescent Zebrafish Screen for Enhancing Hematopoietic Cell Homing.
Stem cell reports
Astuti Y,Kramer AC,Blake AL,Blazar BR,Tolar J,Taisto ME,Lund TC
Published figure using CD184 (CXCR4) monoclonal antibody (Product # 12-9991-82) in Flow Cytometry
Tue Jan 10 00:00:00 UTC 2017
Endogenous TNFα orchestrates the trafficking of neutrophils into and within lymphatic vessels during acute inflammation.
Scientific reports
Arokiasamy S,Zakian C,Dilliway J,Wang W,Nourshargh S,Voisin MB
12-9991 was used in Immunofluorescence to demonstrate a new role for TNFα as a key regulator of neutrophil trafficking into and within the lymphatic system.
Mon Mar 13 00:00:00 UTC 2017
EZH2 enables germinal centre formation through epigenetic silencing of CDKN1A and an Rb-E2F1 feedback loop.
Nature communications
Béguelin W,Rivas MA,Calvo Fernández MT,Teater M,Purwada A,Redmond D,Shen H,Challman MF,Elemento O,Singh A,Melnick AM
12-9991 was used in Flow cytometry/Cell sorting to show EZH2 mediates germinal centre formation through epigenetic silencing of CDKN1A and release of cell cycle checkpoints.
Thu Oct 12 00:00:00 UTC 2017
Single-cell profiling reveals GPCR heterogeneity and functional patterning during neuroinflammation.
JCI insight
Tischner D,Grimm M,Kaur H,Staudenraus D,Carvalho J,Looso M,Günther S,Wanke F,Moos S,Siller N,Breuer J,Schwab N,Zipp F,Waisman A,Kurschus FC,Offermanns S,Wettschureck N
12-9991 was used in Flow cytometry/Cell sorting to identify functionally distinct adaptive GPCR heterogeneity patternation in mice.
Thu Aug 03 00:00:00 UTC 2017
mTOR has distinct functions in generating versus sustaining humoral immunity.
The Journal of clinical investigation
Jones DD,Gaudette BT,Wilmore JR,Chernova I,Bortnick A,Weiss BM,Allman D
12-9991 was used in Flow cytometry/Cell sorting to conclude that mTOR signalling plays critical but diverse roles in early and late phases of antibody responses and plasma cell differentiation.
Tue Nov 01 00:00:00 UTC 2016
Induced Pluripotent Stem Cells derived Muscle Progenitors Effectively Mitigate Muscular Dystrophy through Restoring the Dystrophin Distribution.
Journal of stem cell research & therapy
Cai WF,Huang W,Wang L,Wang JP,Zhang L,Ashraf M,Wu S,Wang Y
Wed Sep 30 00:00:00 UTC 2020
G Protein Coupled Receptor Kinase 3 Regulates Breast Cancer Migration, Invasion, and Metastasis.
PloS one
Billard MJ,Fitzhugh DJ,Parker JS,Brozowski JM,McGinnis MW,Timoshchenko RG,Serafin DS,Lininger R,Klauber-Demore N,Sahagian G,Truong YK,Sassano MF,Serody JS,Tarrant TK
12-9991 was used in Flow cytometry/Cell sorting to demonstrate that G protein coupled receptor kinase 3 dysregulation may play an important part in triple negative breast cancer metastasis.
Fri Aug 26 00:00:00 UTC 2016
Inhibition of insulin-like growth factor receptor/AKT/mammalian target of rapamycin axis targets colorectal cancer stem cells by attenuating mevalonate-isoprenoid pathway in vitro and in vivo.
Oncotarget
Sharon C,Baranwal S,Patel NJ,Rodriguez-Agudo D,Pandak WM,Majumdar AP,Krystal G,Patel BB
Published figure using CD184 (CXCR4) monoclonal antibody (Product # 12-9991-82) in Flow Cytometry
Sat Jun 20 00:00:00 UTC 2015
Intravenous transplantation of very small embryonic like stem cells in treatment of diabetes mellitus.
Avicenna journal of medical biotechnology
Abouzaripour M,Ragerdi Kashani I,Pasbakhsh P,Atlasy N
Published figure using CD184 (CXCR4) monoclonal antibody (Product # 12-9991-82) in Flow Cytometry
Thu Apr 30 00:00:00 UTC 2015
Inhibition of C-terminal binding protein attenuates transcription factor 4 signaling to selectively target colon cancer stem cells.
Cell cycle (Georgetown, Tex.)
Patel J,Baranwal S,Love IM,Patel NJ,Grossman SR,Patel BB
12-9991 was used in Flow cytometry/Cell sorting to present a novel approach for treatment of colorectal cancer focused on targeting of CSCs.
Wed Sep 16 00:00:00 UTC 2015
Efficient definitive endoderm induction from mouse embryonic stem cell adherent cultures: a rapid screening model for differentiation studies.
Stem cell research
Mfopou JK,Geeraerts M,Dejene R,Van Langenhoven S,Aberkane A,Van Grunsven LA,Bouwens L
12-9991 was used in Immunofluorescence to develop a protocol to efficiently induce definitive endoderm cells in mouse embryonic stem cell monolayer cultures.
Wed Jan 01 00:00:00 UTC 2014
Efficient definitive endoderm induction from mouse embryonic stem cell adherent cultures: a rapid screening model for differentiation studies.
Stem cell research
Mfopou JK,Geeraerts M,Dejene R,Van Langenhoven S,Aberkane A,Van Grunsven LA,Bouwens L
12-9991 was used in Immunofluorescence to develop a protocol to efficiently induce definitive endoderm cells in mouse embryonic stem cell monolayer cultures.
Wed Jan 01 00:00:00 UTC 2014
Fibrocytes in the fibrotic lung: altered phenotype detected by flow cytometry.
Frontiers in pharmacology
Reese C,Lee R,Bonner M,Perry B,Heywood J,Silver RM,Tourkina E,Visconti RP,Hoffman S
Published figure using CD184 (CXCR4) monoclonal antibody (Product # 12-9991-82) in Flow Cytometry
Mon Jul 07 00:00:00 UTC 2014
An important role of the SDF-1/CXCR4 axis in chronic skin inflammation.
PloS one
Zgraggen S,Huggenberger R,Kerl K,Detmar M
12-9991 was used in Flow cytometry/Cell sorting to indicate an important role of the SDF-1/CXCR4 axis in skin inflammation and inflammatory angiogenesis.
Mon Feb 02 00:00:00 UTC 2015
Self-recognition of the endothelium enables regulatory T-cell trafficking and defines the kinetics of immune regulation.
Nature communications
Fu H,Kishore M,Gittens B,Wang G,Coe D,Komarowska I,Infante E,Ridley AJ,Cooper D,Perretti M,Marelli-Berg FM
12-9991 was used in Flow cytometry/Cell sorting to explain the mechanisms underlying the powerful migratory capacity of Treg cells and their ability to maintain optimal T cell ratios.
Fri Mar 14 00:00:00 UTC 2014
PP4 is essential for germinal center formation and class switch recombination in mice.
PloS one
Chen MY,Chen YP,Wu MS,Yu GY,Lin WJ,Tan TH,Su YW
12-9991 was used in Flow cytometry/Cell sorting to elucidate the role of the phosphatase PP4 in mature B cells.
Tue Dec 15 00:00:00 UTC 2015
CD73 expression is dynamically regulated in the germinal center and bone marrow plasma cells are diminished in its absence.
PloS one
Conter LJ,Song E,Shlomchik MJ,Tomayko MM
12-9991 was used in Flow cytometry/Cell sorting to identfy CD73-dependent adenosine signaling as a potential novel role for CD73 in humoral immunity.
Thu Jan 15 00:00:00 UTC 2015
Colonic patch and colonic SILT development are independent and differentially regulated events.
Mucosal immunology
Baptista AP,Olivier BJ,Goverse G,Greuter M,Knippenberg M,Kusser K,Domingues RG,Veiga-Fernandes H,Luster AD,Lugering A,Randall TD,Cupedo T,Mebius RE
12-9991 was used in Flow cytometry/Cell sorting to distinguish the different mechanisms responsible for the formation of two distinct colonic lymphoid tissues.
Wed May 01 00:00:00 UTC 2013
Uterine natural killer cells severely decrease in number at gestation day 6 in mice.
Biology of reproduction
Takashima A,Ishikawa F,Kuwabara T,Tanaka Y,Kinoshita T,Ito M,Kakiuchi T
12-9991 was used in Flow cytometry/Cell sorting to study how uterine NK cells or their precursors migrate into the uterus, mature, and produce interferon-gamma to support pregnancy in mice.
Tue Oct 01 00:00:00 UTC 2013
Bortezomib resistance can be reversed by induced expression of plasma cell maturation markers in a mouse in vitro model of multiple myeloma.
PloS one
Stessman HA,Mansoor A,Zhan F,Linden MA,Van Ness B,Baughn LB
12-9991 was used in Flow cytometry/Cell sorting to suggest a novel therapeutic approach for reversing Bz resistance in refractory multiple myeloma.
Tue Aug 05 00:00:00 UTC 2014
Fetal skeletal muscle progenitors have regenerative capacity after intramuscular engraftment in dystrophin deficient mice.
PloS one
Sakai H,Sato T,Sakurai H,Yamamoto T,Hanaoka K,Montarras D,Sehara-Fujisawa A
12-9991 was used in Flow cytometry/Cell sorting to demonstrate the potential of muscle progenitors for muscle repair and cell therapy after degenerative muscle disease.
Tue Dec 24 00:00:00 UTC 2013
Monocytes from Irf5-/- mice have an intrinsic defect in their response to pristane-induced lupus.
Journal of immunology (Baltimore, Md. : 1950)
Yang L,Feng D,Bi X,Stone RC,Barnes BJ
12-9991 was used in Flow cytometry/Cell sorting to examine the mechanism by which loss of Irf5 protects mice from pristane-induced lupus early in disease development.
Mon Oct 01 00:00:00 UTC 2012
Distinct transcriptional signatures of bone marrow-derived C57BL/6 and DBA/2 dendritic leucocytes hosting live Leishmania amazonensis amastigotes.
PLoS neglected tropical diseases
Giraud E,Lecoeur H,Soubigou G,Coppée JY,Milon G,Prina E,Lang T
12-9991 was used in Flow cytometry/Cell sorting to contrast the dendritic cell response to L. amazonensis metacyclic promastigotes in C57BL/6 and DBA/2 mice.
Tue May 14 00:00:00 UTC 2013
Non-invasive imaging provides spatiotemporal information on disease progression and response to therapy in a murine model of multiple myeloma.
PloS one
Riedel SS,Mottok A,Brede C,Bäuerlein CA,Jordán Garrote AL,Ritz M,Mattenheimer K,Rosenwald A,Einsele H,Bogen B,Beilhack A
12-9991 was used in Flow cytometry/Cell sorting to describe a novel MM mouse model that facilitates the convenient, reliable, and sensitive tracking of myeloma cells with bioluminescence imaging.
Tue Jul 02 00:00:00 UTC 2013
Early sensing of Yersinia pestis airway infection by bone marrow cells.
Frontiers in cellular and infection microbiology
Vagima Y,Levy Y,Gur D,Tidhar A,Aftalion M,Abramovich H,Zahavy E,Zauberman A,Flashner Y,Shafferman A,Mamroud E
12-9991 was used in Flow cytometry/Cell sorting to indicate that lung infection by Y. pestis is sensed by BM cells early after infection, although bacterial colonization of the BM occurs at late disease stages.
Wed Dec 11 00:00:00 UTC 2013
Local renal autoantibody production in lupus nephritis.
Journal of the American Society of Nephrology : JASN
Espeli M,Bökers S,Giannico G,Dickinson HA,Bardsley V,Fogo AB,Smith KG
12-9991 was used in Flow cytometry/Cell sorting to identify the kidney as a major reservoir of autoreactive plasma cells in murine models of lupus nephritis that may contribute significantly to disease progression.
Tue Feb 01 00:00:00 UTC 2011
Intramuscular VEGF activates an SDF1-dependent progenitor cell cascade and an SDF1-independent muscle paracrine cascade for cardiac repair.
American journal of physiology. Heart and circulatory physiology
Zisa D,Shabbir A,Mastri M,Taylor T,Aleksic I,McDaniel M,Suzuki G,Lee T
12-9991 was used in Flow cytometry/Cell sorting to investigate two distinct but synergistic cardiac therapeutic mechanisms activated by intramuscular VEGF.
Thu Dec 01 00:00:00 UTC 2011
CD22 expression mediates the regulatory functions of peritoneal B-1a cells during the remission phase of contact hypersensitivity reactions.
Journal of immunology (Baltimore, Md. : 1950)
Nakashima H,Hamaguchi Y,Watanabe R,Ishiura N,Kuwano Y,Okochi H,Takahashi Y,Tamaki K,Sato S,Tedder TF,Fujimoto M
12-9991 was used in Flow cytometry/Cell sorting to clarify the roles of B cells in contact hypersensitivity (CHS), showing that CD22 expression mediates the regulatory functions of peritoneal B-1a cells during the remission phase of CHS.
Sat May 01 00:00:00 UTC 2010
Methyl gallate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.
Journal of immunology (Baltimore, Md. : 1950)
Lee H,Lee H,Kwon Y,Lee JH,Kim J,Shin MK,Kim SH,Bae H
12-9991 was used in Flow cytometry/Cell sorting to study effect of Methyl gallate on Treg cells, leading to reduced tumour growth, due to reversing immune suppression.
Wed Dec 01 00:00:00 UTC 2010
Intramuscular VEGF repairs the failing heart: role of host-derived growth factors and mobilization of progenitor cells.
American journal of physiology. Regulatory, integrative and comparative physiology
Zisa D,Shabbir A,Mastri M,Suzuki G,Lee T
Published figure using CD184 (CXCR4) monoclonal antibody (Product # 12-9991-82) in Flow Cytometry
Sun Nov 01 00:00:00 UTC 2009