Product References
Polycomb Bodies Detection in Murine Fibromuscular Stroma from Skin, Skeletal Muscles, and Aortic Tissues.
Methods in molecular biology (Clifton, N.J.)
Rosti V,Gorini F,Santarelli P,Sarnicola ML,Magnani S,Lanzuolo C
13-5987-82 was used in Flow cytometry/Cell sorting to combine a fluorescence-activated cell sorter (FACS)-based method with an immunofluorescence staining.
Wed May 24 00:00:00 EDT 2023
Machine Learning of Hematopoietic Stem Cell Divisions from Paired Daughter Cell Expression Profiles Reveals Effects of Aging on Self-Renewal.
Cell systems
Arai F,Stumpf PS,Ikushima YM,Hosokawa K,Roch A,Lutolf MP,Suda T,MacArthur BD
13-5987-82 was used in Flow Cytometry to indicate that the balance between intrinsic and extrinsic regulation of stem cell activity alters substantially with age and help explain why stem cell numbers increase through life, yet regenerative potency declines.
Wed Dec 16 00:00:00 EST 2020
A RUNX2 stabilization pathway mediates physiologic and pathologic bone formation.
Nature communications
Kim JM,Yang YS,Park KH,Ge X,Xu R,Li N,Song M,Chun H,Bok S,Charles JF,Filhol-Cochet O,Boldyreff B,Dinter T,Yu PB,Kon N,Gu W,Takarada T,Greenblatt MB,Shim JH
13-5987 was used in Flow cytometry/Cell sorting to demonstrate that CK2-induced phosphorylation of RUNX2 recruits the deubiquitinase HAUSP for the stabilization of RUNX2 and that this pathway is required for physiologic and pathologic bone formation.
Fri May 08 00:00:00 EDT 2020
A RUNX2 stabilization pathway mediates physiologic and pathologic bone formation.
Nature communications
Kim JM,Yang YS,Park KH,Ge X,Xu R,Li N,Song M,Chun H,Bok S,Charles JF,Filhol-Cochet O,Boldyreff B,Dinter T,Yu PB,Kon N,Gu W,Takarada T,Greenblatt MB,Shim JH
13-5987 was used in Flow cytometry/Cell sorting to demonstrate that CK2-induced phosphorylation of RUNX2 recruits the deubiquitinase HAUSP for the stabilization of RUNX2 and that this pathway is required for physiologic and pathologic bone formation.
Fri May 08 00:00:00 EDT 2020
Prrx1 Fibroblasts Represent a Pro-fibrotic Lineage in the Mouse Ventral Dermis.
Cell reports
Leavitt T,Hu MS,Borrelli MR,Januszyk M,Garcia JT,Ransom RC,Mascharak S,desJardins-Park HE,Litzenburger UM,Walmsley GG,Marshall CD,Moore AL,Duoto B,Adem S,Foster DS,Salhotra A,Shen AH,Griffin M,Shen EZ,Barnes LA,Zielins ER,Maan ZN,Wei Y,Chan CKF,Wan DC,Lorenz HP,Chang HY,Gurtner GC,Longaker MT
13-5987-82 was used in Flow Cytometry to identify and characterise a fibroblast subpopulation in the mouse ventral dermis with intrinsic scar-forming potential.
Tue Nov 10 00:00:00 EST 2020
Mesenchymal VEGFA induces aberrant differentiation in heterotopic ossification.
Bone research
Hwang C,Marini S,Huber AK,Stepien DM,Sorkin M,Loder S,Pagani CA,Li J,Visser ND,Vasquez K,Garada MA,Li S,Xu J,Hsu CY,Yu PB,James AW,Mishina Y,Agarwal S,Li J,Levi B
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Flow Cytometry
Sun Oct 17 00:00:00 EDT 2021
Protumoral bone marrow-derived cells migrate via Gβγ-dependent signaling pathways and exhibit a complex repertoire of RhoGEFs.
Journal of cell communication and signaling
Cervantes-Villagrana RD,Color-Aparicio VM,Reyes-Cruz G,Vázquez-Prado J
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Flow Cytometry
Sat Jun 01 00:00:00 EDT 2019
Robust hematopoietic specification requires the ubiquitous Sp1 and Sp3 transcription factors.
Epigenetics & chromatin
Gilmour J,O'Connor L,Middleton CP,Keane P,Gillemans N,Cazier JB,Philipsen S,Bonifer C
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Flow Cytometry
Tue Jun 04 00:00:00 EDT 2019
Isolation and functional assessment of mouse skeletal stem cell lineage.
Nature protocols
Gulati GS,Murphy MP,Marecic O,Lopez M,Brewer RE,Koepke LS,Manjunath A,Ransom RC,Salhotra A,Weissman IL,Longaker MT,Chan CKF
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Macrophages orchestrate breast cancer early dissemination and metastasis.
Nature communications
Linde N,Casanova-Acebes M,Sosa MS,Mortha A,Rahman A,Farias E,Harper K,Tardio E,Reyes Torres I,Jones J,Condeelis J,Merad M,Aguirre-Ghiso JA
13-5987 was used in Flow cytometry/Cell sorting to show that, in pre-malignant lesions, CCL2 produced by cancer cells and myeloid cells attracts CD206+/Tie2+macrophages and induces Wnt-1 upregulation that in turn downregulates E-cadherin junctions in the HER2+early cancer cells.
Tue Jan 02 00:00:00 EST 2018
Neoadjuvant chemotherapy induces breast cancer metastasis through a TMEM-mediated mechanism.
Science translational medicine
Karagiannis GS,Pastoriza JM,Wang Y,Harney AS,Entenberg D,Pignatelli J,Sharma VP,Xue EA,Cheng E,D'Alfonso TM,Jones JG,Anampa J,Rohan TE,Sparano JA,Condeelis JS,Oktay MH
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Immunohistochemistry
Wed Jul 05 00:00:00 EDT 2017
Neoadjuvant chemotherapy induces breast cancer metastasis through a TMEM-mediated mechanism.
Science translational medicine
Karagiannis GS,Pastoriza JM,Wang Y,Harney AS,Entenberg D,Pignatelli J,Sharma VP,Xue EA,Cheng E,D'Alfonso TM,Jones JG,Anampa J,Rohan TE,Sparano JA,Condeelis JS,Oktay MH
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Immunohistochemistry
Wed Jul 05 00:00:00 EDT 2017
The Selective Tie2 Inhibitor Rebastinib Blocks Recruitment and Function of Tie2Hi Macrophages in Breast Cancer and Pancreatic Neuroendocrine Tumors.
Molecular cancer therapeutics
Harney AS,Karagiannis GS,Pignatelli J,Smith BD,Kadioglu E,Wise SC,Hood MM,Kaufman MD,Leary CB,Lu WP,Al-Ani G,Chen X,Entenberg D,Oktay MH,Wang Y,Chun L,De Palma M,Jones JG,Flynn DL,Condeelis JS
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Immunofluorescence
Wed Nov 01 00:00:00 EDT 2017
Pericyte-expressed Tie2 controls angiogenesis and vessel maturation.
Nature communications
Teichert M,Milde L,Holm A,Stanicek L,Gengenbacher N,Savant S,Ruckdeschel T,Hasanov Z,Srivastava K,Hu J,Hertel S,Bartol A,Schlereth K,Augustin HG
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Flow Cytometry
Tue Jul 18 00:00:00 EDT 2017
Primitive erythrocytes are generated from hemogenic endothelial cells.
Scientific reports
Stefanska M,Batta K,Patel R,Florkowska M,Kouskoff V,Lacaud G
13-5987 was used in Immunohistochemistry to demonstrate that both in vivo and in vitro primitive erythrocytes are generated from hemogenic endothelial cells.
Tue Jul 25 00:00:00 EDT 2017
Primitive erythrocytes are generated from hemogenic endothelial cells.
Scientific reports
Stefanska M,Batta K,Patel R,Florkowska M,Kouskoff V,Lacaud G
13-5987 was used in Immunohistochemistry to demonstrate that both in vivo and in vitro primitive erythrocytes are generated from hemogenic endothelial cells.
Tue Jul 25 00:00:00 EDT 2017
Lymphatic Contribution to the Cellular Niche in Heterotopic Ossification.
Annals of surgery
Loder S,Agarwal S,Sorkin M,Breuler C,Li J,Peterson J,Gardenier J,Hsieh HH,Wang SC,Mehrara BJ,Levi B
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
Dynamic Gene Regulatory Networks Drive Hematopoietic Specification and Differentiation.
Developmental cell
Goode DK,Obier N,Vijayabaskar MS,Lie-A-Ling M,Lilly AJ,Hannah R,Lichtinger M,Batta K,Florkowska M,Patel R,Challinor M,Wallace K,Gilmour J,Assi SA,Cauchy P,Hoogenkamp M,Westhead DR,Lacaud G,Kouskoff V,Göttgens B,Bonifer C
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Immunofluorescence
Mon Mar 07 00:00:00 EST 2016
ECM-Dependence of Endothelial Progenitor Cell Features.
Journal of cellular biochemistry
Siavashi V,Nassiri SM,Rahbarghazi R,Vafaei R,Sariri R
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Endoglin potentiates nitric oxide synthesis to enhance definitive hematopoiesis.
Biology open
Nasrallah R,Knezevic K,Thai T,Thomas SR,Göttgens B,Lacaud G,Kouskoff V,Pimanda JE
13598782 was used in flow cytometry to investigate the role of endoglin in the development of hematopoietic cells.
Fri May 15 00:00:00 EDT 2015
The transcription factor Mesp1 interacts with cAMP-responsive element binding protein 1 (Creb1) and coactivates Ets variant 2 (Etv2) gene expression.
The Journal of biological chemistry
Shi X,Zirbes KM,Rasmussen TL,Ferdous A,Garry MG,Koyano-Nakagawa N,Garry DJ
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Immunohistochemistry
Fri Apr 10 00:00:00 EDT 2015
A crucial role for the ubiquitously expressed transcription factor Sp1 at early stages of hematopoietic specification.
Development (Cambridge, England)
Gilmour J,Assi SA,Jaegle U,Kulu D,van de Werken H,Clarke D,Westhead DR,Philipsen S,Bonifer C
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Flow Cytometry
Sun Jun 01 00:00:00 EDT 2014
Cooperative interaction of Etv2 and Gata2 regulates the development of endothelial and hematopoietic lineages.
Developmental biology
Shi X,Richard J,Zirbes KM,Gong W,Lin G,Kyba M,Thomson JA,Koyano-Nakagawa N,Garry DJ
Published figure using CD202b (TIE2) monoclonal antibody (Product # 13-5987-82) in Flow Cytometry
Thu May 15 00:00:00 EDT 2014
Ectopic Runx1 expression rescues Tal-1-deficiency in the generation of primitive and definitive hematopoiesis.
PloS one
Tornack J,Seiler K,Grützkau A,Grün JR,Onodera M,Melchers F,Tsuneto M
13-5987 was used in Flow cytometry/Cell sorting to suggest that Tal-1 expression is needed to express Runx1 in the mesoderm.
Thu Aug 21 00:00:00 EDT 2014
GFI1 and GFI1B control the loss of endothelial identity of hemogenic endothelium during hematopoietic commitment.
Blood
Lancrin C,Mazan M,Stefanska M,Patel R,Lichtinger M,Costa G,Vargel O,Wilson NK,Möröy T,Bonifer C,Göttgens B,Kouskoff V,Lacaud G
13-5987 was used in Flow cytometry/Cell sorting to demonstrate a critical and specific role of the GFI1 transcription factors in the first steps of the process leading to the generation of hematopoietic progenitors.
Thu Jul 12 00:00:00 EDT 2012
GFI1 and GFI1B control the loss of endothelial identity of hemogenic endothelium during hematopoietic commitment.
Blood
Lancrin C,Mazan M,Stefanska M,Patel R,Lichtinger M,Costa G,Vargel O,Wilson NK,Möröy T,Bonifer C,Göttgens B,Kouskoff V,Lacaud G
13-5987 was used in Flow cytometry/Cell sorting to demonstrate a critical and specific role of the GFI1 transcription factors in the first steps of the process leading to the generation of hematopoietic progenitors.
Thu Jul 12 00:00:00 EDT 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Wed Aug 01 00:00:00 EDT 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Wed Aug 01 00:00:00 EDT 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Wed Aug 01 00:00:00 EDT 2012
The notch ligand delta-like 4 regulates multiple stages of early hemato-vascular development.
PloS one
Laranjeiro R,Alcobia I,Neves H,Gomes AC,Saavedra P,Carvalho CC,Duarte A,Cidadão A,Parreira L
13-5987 was used in Flow cytometry/Cell sorting to indicate a role for the notch ligand delta-like 4 in the quantitative regulation of early hemato-vascular precursors.
Thu Aug 30 00:00:00 EDT 2012
Paired immunoglobin-like receptor-B regulates the suppressive function and fate of myeloid-derived suppressor cells.
Immunity
Ma G,Pan PY,Eisenstein S,Divino CM,Lowell CA,Takai T,Chen SH
13-5987 was used in Flow cytometry/Cell sorting to examine the role of paired immunoglobulin-like receptor B (PIR-B) in tumour-induced immunity.
Fri Mar 25 00:00:00 EDT 2011
ER71 directs mesodermal fate decisions during embryogenesis.
Development (Cambridge, England)
Rasmussen TL,Kweon J,Diekmann MA,Belema-Bedada F,Song Q,Bowlin K,Shi X,Ferdous A,Li T,Kyba M,Metzger JM,Koyano-Nakagawa N,Garry DJ
Tue Nov 01 00:00:00 EDT 2011
ER71 directs mesodermal fate decisions during embryogenesis.
Development (Cambridge, England)
Rasmussen TL,Kweon J,Diekmann MA,Belema-Bedada F,Song Q,Bowlin K,Shi X,Ferdous A,Li T,Kyba M,Metzger JM,Koyano-Nakagawa N,Garry DJ
Tue Nov 01 00:00:00 EDT 2011
The haemangioblast generates haematopoietic cells through a haemogenic endothelium stage.
Nature
Lancrin C,Sroczynska P,Stephenson C,Allen T,Kouskoff V,Lacaud G
13-5987 was used in Flow cytometry/Cell sorting to merge the two a priori conflicting theories on the origin of haematopoietic development into a single linear developmental process.
Thu Feb 12 00:00:00 EST 2009
Low levels of tumor necrosis factor alpha increase tumor growth by inducing an endothelial phenotype of monocytes recruited to the tumor site.
Cancer research
Li B,Vincent A,Cates J,Brantley-Sieders DM,Polk DB,Young PP
13-5987 was used in Flow cytometry/Cell sorting to investigate the role of TNF-alpha in endothelial differentiation of myeloid progenitors in tumours.
Thu Jan 01 00:00:00 EST 2009
Mapping of angiogenic markers for targeting of vectors to tumor vascular endothelial cells.
Cancer gene therapy
Tang Y,Borgstrom P,Maynard J,Koziol J,Hu Z,Garen A,Deisseroth A
Sun Apr 01 00:00:00 EDT 2007
Isolation of cardiac cells from E8.5 yolk sac by ALCAM (CD166) expression.
Mechanisms of development
Murakami Y,Hirata H,Miyamoto Y,Nagahashi A,Sawa Y,Jakt M,Asahara T,Kawamata S
13-5987 was used in Flow cytometry/Cell sorting to indicate that cells retaining a potential to differentiate to the cardiac lineage are present in E8.5 yolk sacs and can be isolated as ALCAM high+, Flk-1- cells.
Wed Mar 19 00:00:00 EDT 2008
Differential expression of novel potential regulators in hematopoietic stem cells.
PLoS genetics
Forsberg EC,Prohaska SS,Katzman S,Heffner GC,Stuart JM,Weissman IL
13-5987 was used in Flow cytometry/Cell sorting to compare the expression pattern of three hematopoietic subpopulations in adult mouse bone marrow.
Thu Sep 01 00:00:00 EDT 2005
Postnatal bone marrow stromal cells elicit a potent VEGF-dependent neoangiogenic response in vivo.
Gene therapy
Al-Khaldi A,Eliopoulos N,Martineau D,Lejeune L,Lachapelle K,Galipeau J
13-5987 was used in Flow cytometry/Cell sorting to determine the functional neoangiogenic potential of bone marrow stromal cells in vivo.
Tue Apr 01 00:00:00 EST 2003