Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 1
Immunogen sequence: LHYAKNVRID KVHLFVDQQW LAVRSKSNTN CGGGNHGYNN EFRSMEAIFL AHGPSFKEKT EVEPFEN
The protein encoded by this gene belongs to a series of ectoenzymes that are involved in hydrolysis of extracellular nucleotides. These ectoenzymes possess ATPase and ATP pyrophosphatase activities and are type II transmembrane proteins. Expression of the related rat mRNA has been found in a subset of immature glial cells and in the alimentary tract. The corresponding rat protein has been detected in the pancreas, small intestine, colon, and liver. The human mRNA is expressed in glioma cells, prostate, and uterus. Expression of the human protein has been detected in uterus, basophils, and mast cells. [provided by RefSeq, Jul 2008]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD203c; dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); E-NPP 3; Ectonucleotide pyrophosphatase/phosphodiesterase family member 3; gp130RB13-6; PD-Ibeta; Phosphodiesterase I beta; Phosphodiesterase I/nucleotide pyrophosphatase 3; phosphodiesterase-I beta
Gene Aliases: B10; CD203c; ENPP3; NPP3; PD-IBETA; PDNP3
UniProt ID: (Human) O14638
Entrez Gene ID: (Human) 5169
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support