Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Immunogen sequence: SHAEEVSKFS VCGFANPLPT ESLDCFCPHL QNSTQLEQVN QMLNLTQEEI TATVKVNLPF GRPRVLQKNV DHCLLYHREY VSGFGKAMRM PMWSSYTVPQ LGDTSPLPPT VPDCLRADVR VPPSESQKCS FYLADKNITH GFLYPPASNR TSDSQYDALI TSNLVPMYEE FRKMWDYFHS VLLIKHATER NGVNVVSGPI FDYNYDGHFD APDEITKHLA NTDVPIPTHY FVVLTSCKNK SHTPENCPGW LDVLPFIIPH RPTNVESCPE GKPEALWVEE RFTAHIARVR DVELLTGLDF YQDKVQPVSE ILQLKTYLPT FETTI
The protein encoded by this gene belongs to a series of ectoenzymes that are involved in hydrolysis of extracellular nucleotides. These ectoenzymes possess ATPase and ATP pyrophosphatase activities and are type II transmembrane proteins. Expression of the related rat mRNA has been found in a subset of immature glial cells and in the alimentary tract. The corresponding rat protein has been detected in the pancreas, small intestine, colon, and liver. The human mRNA is expressed in glioma cells, prostate, and uterus. Expression of the human protein has been detected in uterus, basophils, and mast cells. [provided by RefSeq, Jul 2008]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alkaline phosphodiesterase I; CD203c; Dinucleoside polyphosphatase; dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); E-NPP 3; Ectonucleotide pyrophosphatase/phosphodiesterase family member 3; gp130RB13-6; NPPase; Nucleotide diphosphatase; Nucleotide pyrophosphatase; PD-Ibeta; Phosphodiesterase I beta; Phosphodiesterase I/nucleotide pyrophosphatase 3; phosphodiesterase-I beta
Gene Aliases: B10; CD203c; ENPP3; NPP3; PD-IBETA; PDNP3
UniProt ID: (Human) O14638
Entrez Gene ID: (Human) 5169
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support