Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASACPGSD FTSIHSEEEQ LRGLGFRQTR GYKSLAVSKV PSSISQEQSR QDAIYQNLTQ LKAAVGELSE KSKLQEIYQE LTQLKAAVGE LPEKSKLQEI YQELTRLKAA VGELPEKSKL QEIYQELTWL KAAVGELPEK SKMQEIYQEL TRLKAAVGEL PEKSKQQEIY QELTRLKAAV GELPEKSKQQ EIYQELTRLK AAVGELPEKS KQQEIYQELT QLKAAVERLC HPCPWEWTFF QGNCYFMSNS QRNWHDSITA CKEVGAQLVV IKSAEEQNFL QLQSSRSNRF TWMGLSDLNQ EGTWQWVDGS PLLPSFKQYW NRGEPNNVGE EDCAEFSGNG WNDDKCNLAK FWICKKSAAS CSRDEEQFLS PAPATPNPPP A (1-380 aa encoded by BC110615)
This gene encodes a transmembrane receptor and is often referred to as DC-SIGN because of its expression on the surface of dendritic cells and macrophages. The encoded protein is involved in the innate immune system and recognizes numerous evolutionarily divergent pathogens ranging from parasites to viruses with a large impact on public health. The protein is organized into three distinct domains: an N-terminal transmembrane domain, a tandem-repeat neck domain and C-type lectin carbohydrate recognition domain. The extracellular region consisting of the C-type lectin and neck domains has a dual function as a pathogen recognition receptor and a cell adhesion receptor by binding carbohydrate ligands on the surface of microbes and endogenous cells. The neck region is important for homo-oligomerization which allows the receptor to bind multivalent ligands with high avidity. Variations in the number of 23 amino acid repeats in the neck domain of this protein are rare but have a significant impact on ligand binding ability. This gene is closely related in terms of both sequence and function to a neighboring gene (GeneID 10332; often referred to as L-SIGN). DC-SIGN and L-SIGN differ in their ligand-binding properties and distribution. Alternative splicing results in multiple variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type lectin domain family 4 member L; C-type lectin domain family 4, member L; CD209; CD209 antigen; DC-SIGN; Dendritic cell-specific ICAM-3-grabbing non-integrin 1; dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin; dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin; HIV gpl20-binding protein; MGC129965; MGC130443
Gene Aliases: CD209; CDSIGN; CLEC4L; DC-SIGN; DC-SIGN1
UniProt ID: (Human) Q96QQ4
Entrez Gene ID: (Human) 30835
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support