Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MGRAMVARL GLGLLLLALL LPTQIYSSET TTGTSSNSSQ STSNSGLAPN PTNATTKAAG GALQSTASLF VVSLSLLHLY S (1-80 aa encoded by BC007674)
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD24; CD24 antigen; CD24 antigen (small cell lung carcinoma cluster 4 antigen); cluster of differentiation 24; FLJ22950; FLJ43543; heat stable antigen; Heat-stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; MGC75043; Nectadrin; nectadrin heat stable antigen; R13-Ag; Signal transducer CD24; Small cell lung carcinoma cluster 4 antigen; X62 heat stable antigen
Gene Aliases: CD24; CD24A; HSA; Ly-52; nectadrin
UniProt ID: (Human) P25063, (Rat) Q07490, (Mouse) P24807
Entrez Gene ID: (Human) 100133941, (Rat) 25145, (Mouse) 12484
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support