Product References
IL-22 initiates an IL-18-dependent epithelial response circuit to enforce intestinal host defence.
Nature communications
Chiang HY,Lu HH,Sudhakar JN,Chen YW,Shih NS,Weng YT,Shui JW
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Tue Feb 15 00:00:00 EST 2022
Population-wide gene disruption in the murine lung epithelium via AAV-mediated delivery of CRISPR-Cas9 components.
Molecular therapy. Methods & clinical development
Chen H,Durinck S,Patel H,Foreman O,Mesh K,Eastham J,Caothien R,Newman RJ,Roose-Girma M,Darmanis S,Warming S,Lattanzi A,Liang Y,Haley B
25-0242-82 was used in flow cytometry to demonstrate a uniquely effective approach for somatic engineering of the murine lung, which will greatly facilitate the modeling of disease and therapeutic intervention.
Thu Dec 08 00:00:00 EST 2022
FACS and immunomagnetic isolation of early erythroid progenitor cells from mouse fetal liver.
STAR protocols
Braun TW,Kuoch MK,Khandros E,Li H
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Fri Mar 18 00:00:00 EDT 2022
Dormant Nfatc1 reporter-marked basal stem/progenitor cells contribute to mammary lobuloalveoli formation.
iScience
Liu R,Hu H,McNeil M,Xu J,Bi X,Lou P,Guerrero-Juarez CF,Dai X,Plikus MV,Shuai J,Yu Z,Lv C
25-0242-82 was used in Flow cytometry/Cell sorting to show that Nfatc1 expression marks a rare population of mammary epithelial cells with the majority being basal epithelial cells.
Fri Mar 18 00:00:00 EDT 2022
IRE1α drives lung epithelial progenitor dysfunction to establish a niche for pulmonary fibrosis.
American journal of physiology. Lung cellular and molecular physiology
Auyeung VC,Downey MS,Thamsen M,Wenger TA,Backes BJ,Sheppard D,Papa FR
25-0242-82 was used in Flow cytometry/Cell sorting to show that the bifunctional kinase/RNase-IRE1α-a central mediator of the unfolded protein response (UPR) to endoplasmic reticulum (ER) stress is a critical promoter of DATP abundance and function.
Fri Apr 01 00:00:00 EDT 2022
Retrotransposon instability dominates the acquired mutation landscape of mouse induced pluripotent stem cells.
Nature communications
Gerdes P,Lim SM,Ewing AD,Larcombe MR,Chan D,Sanchez-Luque FJ,Walker L,Carleton AL,James C,Knaupp AS,Carreira PE,Nefzger CM,Lister R,Richardson SR,Polo JM,Faulkner GJ
25-0242-82 was used in flow cytometry to reveal the complete spectrum and potential significance of mutations acquired by miPSCs.
Sat Dec 03 00:00:00 EST 2022
Hormone-Responsive BMP Signaling Expands Myoepithelial Cell Lineages and Prevents Alveolar Precocity in Mammary Gland.
Frontiers in cell and developmental biology
Shao C,Lou P,Liu R,Bi X,Li G,Yang X,Sheng X,Xu J,Lv C,Yu Z
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Tue Aug 03 00:00:00 EDT 2021
Hormone-Responsive BMP Signaling Expands Myoepithelial Cell Lineages and Prevents Alveolar Precocity in Mammary Gland.
Frontiers in cell and developmental biology
Shao C,Lou P,Liu R,Bi X,Li G,Yang X,Sheng X,Xu J,Lv C,Yu Z
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Tue Aug 03 00:00:00 EDT 2021
Testosterone metabolites differentially regulate obesogenesis and fat distribution.
Molecular metabolism
Sebo ZL,Rodeheffer MS
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Membrane expression of the estrogen receptor ERα is required for intercellular communications in the mammary epithelium.
Development (Cambridge, England)
Gagniac L,Rusidzé M,Boudou F,Cagnet S,Adlanmerini M,Jeannot P,Gaide N,Giton F,Besson A,Weyl A,Gourdy P,Raymond-Letron I,Arnal JF,Brisken C,Lenfant F
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Wed Mar 11 00:00:00 EDT 2020
Transcriptomic and proteomic signatures of stemness and differentiation in the colon crypt.
Communications biology
Habowski AN,Flesher JL,Bates JM,Tsai CF,Martin K,Zhao R,Ganesan AK,Edwards RA,Shi T,Wiley HS,Shi Y,Hertel KJ,Waterman ML
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Wed Aug 19 00:00:00 EDT 2020
Transcriptomic and proteomic signatures of stemness and differentiation in the colon crypt.
Communications biology
Habowski AN,Flesher JL,Bates JM,Tsai CF,Martin K,Zhao R,Ganesan AK,Edwards RA,Shi T,Wiley HS,Shi Y,Hertel KJ,Waterman ML
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Wed Aug 19 00:00:00 EDT 2020
Analysis of Myeloid Cells in Mouse Tissues with Flow Cytometry.
STAR protocols
Liu Z,Gu Y,Shin A,Zhang S,Ginhoux F
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Fri Jun 19 00:00:00 EDT 2020
An Nfil3-Zeb2-Id2 pathway imposes Irf8 enhancer switching during cDC1 development.
Nature immunology
Bagadia P,Huang X,Liu TT,Durai V,Grajales-Reyes GE,Nitschké M,Modrusan Z,Granja JM,Satpathy AT,Briseño CG,Gargaro M,Iwata A,Kim S,Chang HY,Shaw AS,Murphy TL,Murphy KM
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Sun Sep 01 00:00:00 EDT 2019
Lifelong n-3 Polyunsaturated Fatty Acid Exposure Modulates Size of Mammary Epithelial Cell Populations and Expression of Caveolae Resident Proteins in Fat-1 Mice.
Nutrients
Hillyer LM,Kang JX,Ma DWL
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Tue Oct 15 00:00:00 EDT 2019
Mutations in topoisomerase IIβ result in a B cell immunodeficiency.
Nature communications
Broderick L,Yost S,Li D,McGeough MD,Booshehri LM,Guaderrama M,Brydges SD,Kucharova K,Patel NC,Harr M,Hakonarson H,Zackai E,Cowell IG,Austin CA,Hügle B,Gebauer C,Zhang J,Xu X,Wang J,Croker BA,Frazer KA,Putnam CD,Hoffman HM
25-0242-82 was used in Flow Cytometry to identify mutations in type II topoisomerase as a driver of defective proliferation and survival of B2 cells, leading to a B cell immunodeficiency.
Tue Aug 13 00:00:00 EDT 2019
Adipocyte hypertrophy and lipid dynamics underlie mammary gland remodeling after lactation.
Nature communications
Zwick RK,Rudolph MC,Shook BA,Holtrup B,Roth E,Lei V,Van Keymeulen A,Seewaldt V,Kwei S,Wysolmerski J,Rodeheffer MS,Horsley V
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Tue Sep 04 00:00:00 EDT 2018
Wnt Effector TCF4 Is Dispensable for Wnt Signaling in Human Cancer Cells.
Genes
Hrckulak D,Janeckova L,Lanikova L,Kriz V,Horazna M,Babosova O,Vojtechova M,Galuskova K,Sloncova E,Korinek V
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2018
ATF3 Sustains IL-22-Induced STAT3 Phosphorylation to Maintain Mucosal Immunity Through Inhibiting Phosphatases.
Frontiers in immunology
Glal D,Sudhakar JN,Lu HH,Liu MC,Chiang HY,Liu YC,Cheng CF,Shui JW
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Thu Sep 26 00:00:00 EDT 2019
A Paradoxical Tumor-Suppressor Role for the Rac1 Exchange Factor Vav1 in T Cell Acute Lymphoblastic Leukemia.
Cancer cell
Robles-Valero J,Lorenzo-Martín LF,Menacho-Márquez M,Fernández-Pisonero I,Abad A,Camós M,Toribio ML,Espinosa L,Bigas A,Bustelo XR
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Mon Nov 13 00:00:00 EST 2017
Differential cytokine contributions of perivascular haematopoietic stem cell niches.
Nature cell biology
Asada N,Kunisaki Y,Pierce H,Wang Z,Fernandez NF,Birbrair A,Ma'ayan A,Frenette PS
25-0242 was used in Flow cytometry/Cell sorting to demonstrate that the effects of cytokines regulating haematopoietic stem cell functions are dependent on the producing cell sources.
Wed Mar 01 00:00:00 EST 2017
Adipocyte Accumulation in the Bone Marrow during Obesity and Aging Impairs Stem Cell-Based Hematopoietic and Bone Regeneration.
Cell stem cell
Ambrosi TH,Scialdone A,Graja A,Gohlke S,Jank AM,Bocian C,Woelk L,Fan H,Logan DW,Schürmann A,Saraiva LR,Schulz TJ
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Metabolic shifts in residual breast cancer drive tumor recurrence.
The Journal of clinical investigation
Havas KM,Milchevskaya V,Radic K,Alladin A,Kafkia E,Garcia M,Stolte J,Klaus B,Rotmensz N,Gibson TJ,Burwinkel B,Schneeweiss A,Pruneri G,Patil KR,Sotillo R,Jechlinger M
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Skin Adipocyte Stem Cell Self-Renewal Is Regulated by a PDGFA/AKT-Signaling Axis.
Cell stem cell
Rivera-Gonzalez GC,Shook BA,Andrae J,Holtrup B,Bollag K,Betsholtz C,Rodeheffer MS,Horsley V
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
Casitas B-cell lymphoma (Cbl) proteins protect mammary epithelial cells from proteotoxicity of active c-Src accumulation.
Proceedings of the National Academy of Sciences of the United States of America
Mukhopadhyay C,Triplett A,Bargar T,Heckman C,Wagner KU,Naramura M
25-0242 was used in Flow cytometry/Cell sorting to investigate the protection of MECs from proteotoxic stress.
Tue Dec 20 00:00:00 EST 2016
Batf3 maintains autoactivation of Irf8 for commitment of a CD8α(+) conventional DC clonogenic progenitor.
Nature immunology
Grajales-Reyes GE,Iwata A,Albring J,Wu X,Tussiwand R,Kc W,Kretzer NM,Briseño CG,Durai V,Bagadia P,Haldar M,Schönheit J,Rosenbauer F,Murphy TL,Murphy KM
25-0242 was used in Flow cytometry/Cell sorting to investigate the role of the transcription factors Batf3 and IRF8 in the development of CD8α(+) conventional dendritic cells..
Wed Jul 01 00:00:00 EDT 2015
Rapid depot-specific activation of adipocyte precursor cells at the onset of obesity.
Nature cell biology
Jeffery E,Church CD,Holtrup B,Colman L,Rodeheffer MS
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
The immunosenescence-related gene Zizimin2 is associated with early bone marrow B cell development and marginal zone B cell formation.
Immunity & ageing : I & A
Matsuda T,Yanase S,Takaoka A,Maruyama M
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Mon Mar 02 00:00:00 EST 2015
Dynamic regulation of CD24 expression and release of CD24-containing microvesicles in immature B cells in response to CD24 engagement.
Immunology
Ayre DC,Elstner M,Smith NC,Moores ES,Hogan AM,Christian SL
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2015
Dynamic regulation of CD24 expression and release of CD24-containing microvesicles in immature B cells in response to CD24 engagement.
Immunology
Ayre DC,Elstner M,Smith NC,Moores ES,Hogan AM,Christian SL
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2015
The Transcriptional Repressor ZNF503/Zeppo2 Promotes Mammary Epithelial Cell Proliferation and Enhances Cell Invasion.
The Journal of biological chemistry
Shahi P,Slorach EM,Wang CY,Chou J,Lu A,Ruderisch A,Werb Z
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Fri Feb 06 00:00:00 EST 2015
FGF ligands of the postnatal mammary stroma regulate distinct aspects of epithelial morphogenesis.
Development (Cambridge, England)
Zhang X,Martinez D,Koledova Z,Qiao G,Streuli CH,Lu P
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
Rhof promotes murine marginal zone B cell development.
Nagoya journal of medical science
Kishimoto M,Matsuda T,Yanase S,Katsumi A,Suzuki N,Ikejiri M,Takagi A,Ikawa M,Kojima T,Kunishima S,Kiyoi H,Naoe T,Matsushita T,Maruyama M
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Fri Aug 01 00:00:00 EDT 2014
Prospective identification of functionally distinct stem cells and neurosphere-initiating cells in adult mouse forebrain.
eLife
Mich JK,Signer RA,Nakada D,Pineda A,Burgess RJ,Vue TY,Johnson JE,Morrison SJ
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Wed May 07 00:00:00 EDT 2014
Leukocyte population dynamics and detection of IL-9 as a major cytokine at the mouse fetal-maternal interface.
PloS one
Habbeddine M,Verbeke P,Karaz S,Bobé P,Kanellopoulos-Langevin C
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Wed Jun 10 00:00:00 EDT 2015
Characterization of the adipocyte cellular lineage in vivo.
Nature cell biology
Berry R,Rodeheffer MS
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
Evidence for a multipotent mammary progenitor with pregnancy-specific activity.
Breast cancer research : BCR
Kaanta AS,Virtanen C,Selfors LM,Brugge JS,Neel BG
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Mon Dec 08 00:00:00 EST 2014
FLT3-ITDs instruct a myeloid differentiation and transformation bias in lymphomyeloid multipotent progenitors.
Cell reports
Mead AJ,Kharazi S,Atkinson D,Macaulay I,Pecquet C,Loughran S,Lutteropp M,Woll P,Chowdhury O,Luc S,Buza-Vidas N,Ferry H,Clark SA,Goardon N,Vyas P,Constantinescu SN,Sitnicka E,Nerlov C,Jacobsen SE
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Thu Jun 27 00:00:00 EDT 2013
Lunatic fringe deficiency cooperates with the Met/Caveolin gene amplicon to induce basal-like breast cancer.
Cancer cell
Xu K,Usary J,Kousis PC,Prat A,Wang DY,Adams JR,Wang W,Loch AJ,Deng T,Zhao W,Cardiff RD,Yoon K,Gaiano N,Ling V,Beyene J,Zacksenhaus E,Gridley T,Leong WL,Guidos CJ,Perou CM,Egan SE
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Tue May 15 00:00:00 EDT 2012
Restricted expression of new guanine nucleotide exchange factor Zizimin2 in aged acquired immune system.
Nagoya journal of medical science
Jia Y,Sakabe I,Matsuda T,Hayakawa T,Maruyama M
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2012
Stem cell marker prominin-1 regulates branching morphogenesis, but not regenerative capacity, in the mammary gland.
Developmental dynamics : an official publication of the American Association of Anatomists
Anderson LH,Boulanger CA,Smith GH,Carmeliet P,Watson CJ
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2011
A multifunctional 3D co-culture system for studies of mammary tissue morphogenesis and stem cell biology.
PloS one
Campbell JJ,Davidenko N,Caffarel MM,Cameron RE,Watson CJ
Published figure using CD24 monoclonal antibody (Product # 25-0242-82) in Flow Cytometry
Mon Mar 05 00:00:00 EST 2012