Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
FIGURE: 1 / 7
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with Mouse, Sheep and Pig
CD3z complex is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules, and this association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. Diseases associated with CD3z include Immunodeficiency 25 and T-B+ Severe Combined Immunodeficiency Due To Cd3delta/Cd3epsilon/Cd3zeta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD247; CD247 antigen, zeta subunit; CD3-epsilon; CD3e antigen; CD3Z antigen, zeta polypeptide (TiT3 complex); CD3zeta chain; FLJ18683; T-cell antigen receptor complex, zeta subunit of CD3; T-cell receptor T3 zeta chain; T-cell surface glycoprotein CD3 zeta chain; T3E; TCR zeta chain; TCRE
Gene Aliases: CD247; CD3-ZETA; CD3H; CD3Q; CD3Z; IMD25; T3Z; TCRZ
UniProt ID: (Human) P20963
Entrez Gene ID: (Human) 919
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support