Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLLRLLLAWA AAGPTLGQDP WAAEPRAACG PSSCYALFPR RRTFLEAWRA CRELGGDLAT PRTPEEAQRV DSLVGAGPAS RLLWIGLQRQ ARQCQLQRPL RGFTWTTGDQ DTAFTNWAQP ASGGPCPAQR CVALEASGEH RWLEGSCTLA VDGYLCQFGF EGACPALQDE AGQAGPAVYT TPFHLVSTEF EWLPFGSVAA VQCQAGRGAS LLCVKQPEGG VGWSRAGPLC LGTGCSPDNG GCEHECVEEV DGHVSCRCTE GFRLAADGRS CEDPCAQAPC EQQCEPGGPQ GYSCHCRLGF RPAEDDPHRC VDTDECQIAG VCQQMCVNYV GGFECYCSEG HELEADGISC SPAGAMGAQA SQDLGDELLD DGEDEEDEDE AWKAFNGGWT EMPGILWMEP TQPPDFALAY RPSFPEDREP QIPYPEPTWP PPLSAPRVPY HSSVLSVTRP VVVSATRPTL PSAHQPPVIP ATHPALSRDH QIPVIAANYP DLPSAYQPGI LSVSHSAQPP AHQPPMISTK YPELFPAHQS PMFPDTRVAG TQTTTHLPGI PPNHAPLVTT LGAQLPPQAP DALVLRTQAT QLPIIPTAQP SLTTTSRSPV SPAHQISVPA ATQPAALPTL LPSQSPTNQT SPISPTHPHS KAPQIPREDG PSPKLALWLP SPAPTAAPTA LGEAGLAEHS QRDDRWLLVA LLVPTCVFLV VLLALGIVYC TRCGPHAPNK RITDCYRWVI HAGSKSPTEP MPPRGSLTGV QTCRTSV
CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2610111G01Rik; CD164 sialomucin-like 1; CD248; CD248 antigen, endosialin; CD248 molecule, endosialin; Endosialin; MGC119478; MGC119479; Tumor endothelial marker 1
Gene Aliases: CD164L1; CD248; TEM1
UniProt ID: (Human) Q9HCU0
Entrez Gene ID: (Human) 57124
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support