Product References
Advanced immunophenotyping: A powerful tool for immune profiling, drug screening, and a personalized treatment approach.
Frontiers in immunology
Preglej T,Brinkmann M,Steiner G,Aletaha D,Göschl L,Bonelli M
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Tue Apr 11 00:00:00 EDT 2023
Rotenone aggravates PD-like pathology in A53T mutant human α-synuclein transgenic mice in an age-dependent manner.
Frontiers in aging neuroscience
Chen AD,Cao JX,Chen HC,Du HL,Xi XX,Sun J,Yin J,Jing YH,Gao LP
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Fri Aug 26 00:00:00 EDT 2022
Notch4 signaling limits regulatory T-cell-mediated tissue repair and promotes severe lung inflammation in viral infections.
Immunity
Harb H,Benamar M,Lai PS,Contini P,Griffith JW,Crestani E,Schmitz-Abe K,Chen Q,Fong J,Marri L,Filaci G,Del Zotto G,Pishesha N,Kolifrath S,Broggi A,Ghosh S,Gelmez MY,Oktelik FB,Cetin EA,Kiykim A,Kose M,Wang Z,Cui Y,Yu XG,Li JZ,Berra L,Stephen-Victor E,Charbonnier LM,Zanoni I,Ploegh H,Deniz G,De Palma R,Chatila TA
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Tue Jun 08 00:00:00 EDT 2021
FABP4 facilitates inflammasome activation to induce the Treg/Th17 imbalance in preeclampsia via forming a positive feedback with IL-17A.
Molecular therapy. Nucleic acids
Chang GP,Yang XL,Liu W,Lin S,Yang SL,Zhao MY
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Fri Jun 04 00:00:00 EDT 2021
Soluble fibrinogen‑like protein 2 levels are decreased in patients with ischemic heart failure and associated with cardiac function.
Molecular medicine reports
You Y,Huang S,Liu H,Fan C,Liu K,Wang Z
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2021
Imbalance between T helper 1 and regulatory T cells plays a detrimental role in experimental Parkinson's disease in mice.
The Journal of international medical research
Li W,Luo Y,Xu H,Ma Q,Yao Q
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
T cell receptor-dependent S-acylation of ZAP-70 controls activation of T cells.
The Journal of biological chemistry
Tewari R,Shayahati B,Fan Y,Akimzhanov AM
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Thu Aug 26 00:00:00 EDT 2021
Identification of celastrol as a novel HIV-1 latency reversal agent by an image-based screen.
PloS one
Liu H,Hu PW,Dubrulle J,Stossi F,Nikolai BC,Mancini MA,Rice AP
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Tue Sep 14 00:00:00 EDT 2021
Numerical optimization of microfluidic vortex shedding for genome editing T cells with Cas9.
Scientific reports
Jarrell JA,Sytsma BJ,Wilson LH,Pan FL,Lau KHWJ,Kirby GTS,Lievano AA,Pawell RS
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Thu Jun 03 00:00:00 EDT 2021
Human endoglin-CD3 bispecific T cell engager antibody induces anti-tumor effect in vivo.
Theranostics
Zhong L,Shi W,Gan L,Liu X,Huo Y,Wu P,Zhang Z,Wu T,Peng H,Huang Y,Zhao Y,Yuan Y,Deng Z,Tang H
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Wed Jul 28 00:00:00 EDT 2021
Immunophenotype of Gastric Tumors Unveils a Pleiotropic Role of Regulatory T Cells in Tumor Development.
Cancers
Rocha S,Basto AP,Ijsselsteijn ME,Teles SP,Azevedo MM,Gonçalves G,Gullo I,Almeida GM,Maqueda JJ,Oliveira MI,Carneiro F,Barata JT,Graça L,de Miranda NFCC,Carvalho J,Oliveira C
25-0259-42 was used in Flow Cytometry to explore the impact of regulatory T cell (Tregs) in gastric cancers (GCs) with distinct histology, and whether Tregs can directly influence tumor cell behavior and GC development.
Sat Jan 23 00:00:00 EST 2021
Epinephrine Production in Th17 Cells and Experimental Autoimmune Encephalitis.
Frontiers in immunology
Yang P,Tian H,Zou YR,Chambon P,Ichinose H,Honig G,Diamond B,Kim SJ
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Mon Jul 05 00:00:00 EDT 2021
Expansion of Group 2 Innate Lymphoid Cells in Patients with End-Stage Renal Disease and Their Clinical Significance.
Journal of immunology (Baltimore, Md. : 1950)
Liu GY,Deng XH,Li X,Cao YJ,Xing YF,Zhou P,Lei AH,Yang Q,Deng K,Zhang H,Zhou J
25-0259-42 was used in Flow Cytometry to indicate that ILC2s may play a protective role in ESRD.
Wed Jul 01 00:00:00 EDT 2020
Potential of neutrophil to lymphocyte ratio in predicting sustained remission in rheumatoid arthritis compared to other immune activation markers.
The Indian journal of medical research
Chandrashekara S,Lingaraju DC,Renuka P,Anupama KR
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2020
Involvement of natural killer cells in the pathogenesis of endometriosis in patients with pelvic pain.
The Journal of international medical research
He J,Xu Y,Yi M,Gu C,Zhu Y,Hu G
25-0259 was used in Flow cytometry/Cell sorting to investigate the involvement of immune cells in the pathogenesis of endometriosis progression by analyzing blood samples from patients with stable disease and those with pelvic pain.
Wed Jul 01 00:00:00 EDT 2020
Involvement of natural killer cells in the pathogenesis of endometriosis in patients with pelvic pain.
The Journal of international medical research
He J,Xu Y,Yi M,Gu C,Zhu Y,Hu G
25-0259 was used in Flow cytometry/Cell sorting to investigate the involvement of immune cells in the pathogenesis of endometriosis progression by analyzing blood samples from patients with stable disease and those with pelvic pain.
Wed Jul 01 00:00:00 EDT 2020
Th17 reprogramming of T cells in systemic juvenile idiopathic arthritis.
JCI insight
Henderson LA,Hoyt KJ,Lee PY,Rao DA,Jonsson AH,Nguyen JP,Rutherford K,Julé AM,Charbonnier LM,Case S,Chang MH,Cohen EM,Dedeoglu F,Fuhlbrigge RC,Halyabar O,Hazen MM,Janssen E,Kim S,Lo J,Lo MS,Meidan E,Son MBF,Sundel RP,Stoll ML,Nusbaum C,Lederer JA,Chatila TA,Nigrovic PA
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Thu Mar 26 00:00:00 EDT 2020
Th2 Biased Immunity With Altered B Cell Profiles in Circulation of Patients With Sporotrichosis Caused by Sporothrix globosa.
Frontiers in immunology
Zu J,Yao L,Song Y,Cui Y,Guan M,Chen R,Zhen Y,Li S
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Mon May 31 00:00:00 EDT 2021
Functionally specialized human CD4+ T-cell subsets express physicochemically distinct TCRs.
eLife
Kasatskaya SA,Ladell K,Egorov ES,Miners KL,Davydov AN,Metsger M,Staroverov DB,Matveyshina EK,Shagina IA,Mamedov IZ,Izraelson M,Shelyakin PV,Britanova OV,Price DA,Chudakov DM
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Tue Dec 08 00:00:00 EST 2020
Effects of Desflurane and Sevoflurane anesthesia on regulatory T cells in patients undergoing living donor kidney transplantation: a randomized intervention trial.
BMC anesthesiology
Chutipongtanate A,Prukviwat S,Pongsakul N,Srisala S,Kamanee N,Arpornsujaritkun N,Gesprasert G,Apiwattanakul N,Hongeng S,Ittichaikulthol W,Sumethkul V,Chutipongtanate S
25-0259 was used in Flow cytometry/Cell sorting to investigate effects of desflurane and sevoflurane anesthesia on peripheral blood Tregs induction in patients undergoing living donor kidney transplantation.
Thu Aug 27 00:00:00 EDT 2020
Tumors establish resistance to immunotherapy by regulating Treg recruitment via CCR4.
Journal for immunotherapy of cancer
Marshall LA,Marubayashi S,Jorapur A,Jacobson S,Zibinsky M,Robles O,Hu DX,Jackson JJ,Pookot D,Sanchez J,Brovarney M,Wadsworth A,Chian D,Wustrow D,Kassner PD,Cutler G,Wong B,Brockstedt DG,Talay O
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Sun Nov 01 00:00:00 EDT 2020
Loading of Primary Human T Lymphocytes with Citrate-Coated Superparamagnetic Iron Oxide Nanoparticles Does Not Impair Their Activation after Polyclonal Stimulation.
Cells
Mühlberger M,Unterweger H,Band J,Lehmann C,Heger L,Dudziak D,Alexiou C,Lee G,Janko C
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Sat Feb 01 00:00:00 EST 2020
T-Lymphocyte Subset Distribution and Activity in Patients With Glaucoma.
Investigative ophthalmology & visual science
Yang X,Zeng Q,Göktas E,Gopal K,Al-Aswad L,Blumberg DM,Cioffi GA,Liebmann JM,Tezel G
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2019
Transcriptional down-regulation of ccr5 in a subset of HIV+ controllers and their family members.
eLife
Gonzalo-Gil E,Rapuano PB,Ikediobi U,Leibowitz R,Mehta S,Coskun AK,Porterfield JZ,Lampkin TD,Marconi VC,Rimland D,Walker BD,Deeks S,Sutton RE
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Tue Apr 09 00:00:00 EDT 2019
A probiotic modulates the microbiome and immunity in multiple sclerosis.
Annals of neurology
Tankou SK,Regev K,Healy BC,Tjon E,Laghi L,Cox LM,Kivisäkk P,Pierre IV,Hrishikesh L,Gandhi R,Cook S,Glanz B,Stankiewicz J,Weiner HL
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Secretory High-Mobility Group Box 1 Protein Affects Regulatory T Cell Differentiation in Neuroblastoma Microenvironment In Vitro.
Journal of oncology
Vanichapol T,Chiangjong W,Panachan J,Anurathapan U,Chutipongtanate S,Hongeng S
25-0259 was used in Flow cytometry/Cell sorting to elucidate the role of the neuroblastoma microenvironment in promoting regulatory T cell phenotype.
Thu Oct 05 00:00:00 EDT 2023
DNA Methyltransferase Inhibition Promotes Th1 Polarization in Human CD4+CD25high FOXP3+ Regulatory T Cells but Does Not Affect Their Suppressive Capacity.
Journal of immunology research
Landman S,Cruijsen M,Urbano PCM,Huls G,van Erp PEJ,van Rijssen E,Joosten I,Koenen HJPM
25-0259 was used in Flow cytometry/Cell sorting to examine the in vitro effect of derivative 5-aza-2'-deoxycytidine on the stability and function of FACS-sorted human naturally occurring CD4+CD25high FOXP3+ regulatory T cells.
Thu Oct 11 00:00:00 EDT 2018
Intralymphatic Glutamic Acid Decarboxylase-Alum Administration Induced Th2-Like-Specific Immunomodulation in Responder Patients: A Pilot Clinical Trial in Type 1 Diabetes.
Journal of diabetes research
Tavira B,Barcenilla H,Wahlberg J,Achenbach P,Ludvigsson J,Casas R
25-0259 was used in Flow cytometry/Cell sorting to compare the immunomodulatory effect of giving GAD-alum directly into lymph nodes versus that induced by subcutaneous administration.
Tue Dec 11 00:00:00 EST 2018
CD4+CD45RA-FOXP3low Regulatory T Cells as Potential Biomarkers of Disease Activity in Systemic Lupus Erythematosus Brazilian Patients.
BioMed research international
Silva-Neta HL,Brelaz-de-Castro MCA,Chagas MBO,Mariz HA,de Arruda RG,de Vasconcelos VF,Pereira MC,Romano A,Pitta IR,Marques CDL,Duarte ALBP,Rêgo MJBM,Pitta MGR
25-0259 was used in Flow cytometry/Cell sorting to demonstrate Foxp3+nontreg cells as new biomarkers in the search of an effective therapeutic strategy in SLE.
Fri Jan 11 00:00:00 EST 2019
The Effects of High Mobility Group Box-1 Protein on Peripheral Treg/Th17 Balance in Patients with Atherosclerosis.
Acta Cardiologica Sinica
Ding JW,Zhou T,Zheng XX,Wang XA,Tong XH,Luo CY,Zhang ZQ,Yu B
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2018
Disruption of PD-1 Enhanced the Anti-tumor Activity of Chimeric Antigen Receptor T Cells Against Hepatocellular Carcinoma.
Frontiers in pharmacology
Guo X,Jiang H,Shi B,Zhou M,Zhang H,Shi Z,Du G,Luo H,Wu X,Wang Y,Sun R,Li Z
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Thu Sep 28 00:00:00 EDT 2023
Immune evasion mediated by PD-L1 on glioblastoma-derived extracellular vesicles.
Science advances
Ricklefs FL,Alayo Q,Krenzlin H,Mahmoud AB,Speranza MC,Nakashima H,Hayes JL,Lee K,Balaj L,Passaro C,Rooj AK,Krasemann S,Carter BS,Chen CC,Steed T,Treiber J,Rodig S,Yang K,Nakano I,Lee H,Weissleder R,Breakefield XO,Godlewski J,Westphal M,Lamszus K,Freeman GJ,Bronisz A,Lawler SE,Chiocca EA
25-0259 was used in Flow cytometry/Cell sorting to investigate whether glioblastoma extracellular vesicles are important mediators of immunosuppression and whether programmed death ligand-1 could play a role.
Thu Mar 01 00:00:00 EST 2018
Staphylococcus aureus PSM Peptides Modulate Human Monocyte-Derived Dendritic Cells to Prime Regulatory T Cells.
Frontiers in immunology
Richardson JR,Armbruster NS,Günter M,Henes J,Autenrieth SE
25-0259 was used in Flow cytometry/Cell sorting to study the effect of PSMα3 on the maturation, cytokine production, antigen uptake, and T cell stimulatory capacity of human monocyte-derived DCs treated simultaneously with either LPS(TLR4 ligand) or Sa cell lysate (TLR2 ligand).
Thu Oct 03 00:00:00 EDT 2019
Increased serum concentrations of IL-1 beta, IL-21 and Th17 cells in overweight patients with rheumatoid arthritis.
Arthritis research & therapy
Shoda H,Nagafuchi Y,Tsuchida Y,Sakurai K,Sumitomo S,Fujio K,Yamamoto K
25-0259 was used in Flow cytometry/Cell sorting to show individuals with BMI > 25 and rheumatoid arthritis had heightened concentrations of IL-1 beta, IL-21 and Th17 cells.
Wed May 31 00:00:00 EDT 2017
Alteration of Th17 and Foxp3+ regulatory T cells in patients with unexplained recurrent spontaneous abortion before and after the therapy of hCG combined with immunoglobulin.
Experimental and therapeutic medicine
Sha J,Liu F,Zhai J,Liu X,Zhang Q,Zhang B
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
Hormonal vitamin D up-regulates tissue-specific PD-L1 and PD-L2 surface glycoprotein expression in humans but not mice.
The Journal of biological chemistry
Dimitrov V,Bouttier M,Boukhaled G,Salehi-Tabar R,Avramescu RG,Memari B,Hasaj B,Lukacs GL,Krawczyk CM,White JH
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Fri Dec 15 00:00:00 EST 2017
Combinations of isoform-targeted histone deacetylase inhibitors and bryostatin analogues display remarkable potency to activate latent HIV without global T-cell activation.
Scientific reports
Albert BJ,Niu A,Ramani R,Marshall GR,Wender PA,Williams RM,Ratner L,Barnes AB,Kyei GB
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Mon Aug 07 00:00:00 EDT 2017
TIGIT expressing CD4+T cells represent a tumor-supportive T cell subset in chronic lymphocytic leukemia.
Oncoimmunology
Catakovic K,Gassner FJ,Ratswohl C,Zaborsky N,Rebhandl S,Schubert M,Steiner M,Gutjahr JC,Pleyer L,Egle A,Hartmann TN,Greil R,Geisberger R
25-0259 was used in Flow cytometry/Cell sorting to reveal that TIGIT+CD4+T cells provide a supportive microenvironment for CLL cells.
Sat Jan 29 00:00:00 EST 2022
Selective blockade of CD28 on human T cells facilitates regulation of alloimmune responses.
JCI insight
Zaitsu M,Issa F,Hester J,Vanhove B,Wood KJ
25-0259 was used in Flow cytometry/Cell sorting to show that a nonactivating monovalent anti-CD28 that spares CTLA4 signaling is an effective immunosuppressant in a clinically relevant humanized mouse transplant model.
Thu Oct 05 00:00:00 EDT 2017
Azacytidine prevents experimental xenogeneic graft-versus-host disease without abrogating graft-versus-leukemia effects.
Oncoimmunology
Ehx G,Fransolet G,de Leval L,D'Hondt S,Lucas S,Hannon M,Delens L,Dubois S,Drion P,Beguin Y,Humblet-Baron S,Baron F
25-0259 was used in Flow cytometry/Cell sorting to investigate the impact of 5-azacytidine on xenogeneic graft-vs.-host disease and graft-vs.-leukaemia effects in a humanised murine model of transplantation.
Tue Jan 12 00:00:00 EST 2021
Toll-like receptor 2 induced cytotoxic T-lymphocyte-associated protein 4 regulates Aspergillus-induced regulatory T-cells with pro-inflammatory characteristics.
Scientific reports
Raijmakers RPH,Sprenkeler EGG,Aleva FE,Jacobs CWM,Kanneganti TD,Joosten LAB,van de Veerdonk FL,Gresnigt MS
25-0259 was used in Flow cytometry/Cell sorting to suggest a role for regulatory T-cells with a pro-inflammatory TH17-like phenotype in Aspergillus-associated immunopathology, and identifies key players, i.e. TLR2 and CTLA4, involved in this mechanism.
Wed Sep 13 00:00:00 EDT 2017
Alloantigen-specific regulatory T cells generated with a chimeric antigen receptor.
The Journal of clinical investigation
MacDonald KG,Hoeppli RE,Huang Q,Gillies J,Luciani DS,Orban PC,Broady R,Levings MK
25-0259 was used in Flow cytometry/Cell sorting to investigate alloantigen-specific regulatory T cells generated with a chimeric antigen receptor.
Fri Apr 01 00:00:00 EDT 2016
Human Head and Neck Squamous Cell Carcinoma-Associated Semaphorin 4D Induces Expansion of Myeloid-Derived Suppressor Cells.
Journal of immunology (Baltimore, Md. : 1950)
Younis RH,Han KL,Webb TJ
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2016
Lin28b Regulates Fetal Regulatory T Cell Differentiation through Modulation of TGF-β Signaling.
Journal of immunology (Baltimore, Md. : 1950)
Bronevetsky Y,Burt TD,McCune JM
25-0259 was used in Flow cytometry/Cell sorting to determine whether loss of Lin28b expression in foetal T cells leads to increased mature let-7, which causes decreased expression of TGF-ΒRI, TGF-ΒRIII, and SMAD2 proteins.
Thu Dec 01 00:00:00 EST 2016
Equilibrium of Treg/Th17 cells of peripheral blood in syphilitic patients with sero-resistance.
Experimental and therapeutic medicine
Zhao J,Ma J,Zhang X,Li Q,Yang X
Published figure using CD25 monoclonal antibody (Product # 25-0259-42) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2016
Immunophenotyping of rheumatoid arthritis reveals a linkage between HLA-DRB1 genotype, CXCR4 expression on memory CD4(+) T cells, and disease activity.
Scientific reports
Nagafuchi Y,Shoda H,Sumitomo S,Nakachi S,Kato R,Tsuchida Y,Tsuchiya H,Sakurai K,Hanata N,Tateishi S,Kanda H,Ishigaki K,Okada Y,Suzuki A,Kochi Y,Fujio K,Yamamoto K
25-0259 was used in Flow cytometry/Cell sorting to perform 24-subset immunophenotyping of peripheral blood mononuclear cells in HLA-DRB1-genotyped rheumatoid arthritis patients and healthy donors.
Thu Jul 07 00:00:00 EDT 2016
A Systematic Study of the Effect of Different Molecular Weights of Hyaluronic Acid on Mesenchymal Stromal Cell-Mediated Immunomodulation.
PloS one
Gómez-Aristizábal A,Kim KP,Viswanathan S
25-0259 was used in Flow cytometry/Cell sorting to investigate the combined mechanism of action of MSCs and hyaluronic acid.
Wed Jul 06 00:00:00 EDT 2016
PD-L1 mediated the differentiation of tumor-infiltrating CD19+ B lymphocytes and T cells in Invasive breast cancer.
Oncoimmunology
Guan H,Lan Y,Wan Y,Wang Q,Wang C,Xu L,Chen Y,Liu W,Zhang X,Li Y,Gu Y,Wang Z,Xie F
25-0259 was used in Flow cytometry/Cell sorting to investigate the role of B cells in human breast cancer etiology.
Mon Feb 01 00:00:00 EST 2016
Agonist anti-GITR monoclonal antibody and stereotactic radiation induce immune-mediated survival advantage in murine intracranial glioma.
Journal for immunotherapy of cancer
Patel MA,Kim JE,Theodros D,Tam A,Velarde E,Kochel CM,Francica B,Nirschl TR,Ghasemzadeh A,Mathios D,Harris-Bookman S,Jackson CC,Jackson C,Ye X,Tran PT,Tyler B,Coric V,Selby M,Brem H,Drake CG,Pardoll DM,Lim M
25-0259 was used in Flow cytometry/Cell sorting to provide preclinical evidence for the use of anti-GITR (1) non-depleting antibodies in combination with SRS in GBM.
Wed May 18 00:00:00 EDT 2016
Hyperreactive onchocerciasis is characterized by a combination of Th17-Th2 immune responses and reduced regulatory T cells.
PLoS neglected tropical diseases
Katawa G,Layland LE,Debrah AY,von Horn C,Batsa L,Kwarteng A,Arriens S,W Taylor D,Specht S,Hoerauf A,Adjobimey T
25-0259 was used in Flow cytometry/Cell sorting to provide initial evidence that elevated frequencies of Th17 and Th2 cells form part of the immune network instigating the development of severe onchocerciasis.
Thu Jan 01 00:00:00 EST 2015
The number of CCR5 expressing CD4+ T lymphocytes is lower in HIV-infected long-term non-progressors with viral control compared to normal progressors: a cross-sectional study.
BMC infectious diseases
Meijerink H,Indrati AR,van Crevel R,Joosten I,Koenen H,van der Ven AJ
25-0259 was used in Flow cytometry/Cell sorting to determine levels of CCR5 and CXCR4 expression among CD4(+) and CD8(+) lymphocyte subsets in HIV-infected long-term non-progressors.
Sat Dec 13 00:00:00 EST 2014
Elevated frequencies of highly activated CD4+ T cells in HIV+ patients developing immune reconstitution inflammatory syndrome.
Blood
Antonelli LR,Mahnke Y,Hodge JN,Porter BO,Barber DL,DerSimonian R,Greenwald JH,Roby G,Mican J,Sher A,Roederer M,Sereti I
25-0259 was used in Flow cytometry/Cell sorting to indicate that IRIS appears to be a predominantly CD4-mediated phenomenon.
Thu Nov 11 00:00:00 EST 2010
Human Th1 cells that express CD300a are polyfunctional and after stimulation up-regulate the T-box transcription factor eomesodermin.
PloS one
Narayanan S,Silva R,Peruzzi G,Alvarez Y,Simhadri VR,Debell K,Coligan JE,Borrego F
25-0259 was used in Flow cytometry/Cell sorting to investigate a subset within the CD4 T cell population defined by expression of the immunomodulatory receptor CD300a.
Thu May 13 00:00:00 EDT 2010
Fc receptor-like 3 protein expressed on IL-2 nonresponsive subset of human regulatory T cells.
Journal of immunology (Baltimore, Md. : 1950)
Nagata S,Ise T,Pastan I
25-0259 was used in Flow cytometry/Cell sorting to study the effects of Foxp3 and IL-2 on FCRL3-/+ nTreg cell subsets.
Mon Jun 15 00:00:00 EDT 2009
Rhesus monkey immature monocyte-derived dendritic cells generate alloantigen-specific regulatory T cells from circulating CD4+CD127-/lo T cells.
Transplantation
Zahorchak AF,Raimondi G,Thomson AW
25-0259 was used in Flow cytometry/Cell sorting to exhibit a novel method for the generation of non-human primate regulatory T cells with alloantigen specificity.
Sun Nov 15 00:00:00 EST 2009
Immune cellular parameters of leprosy and human immunodeficiency virus-1 co-infected subjects.
Immunology
Carvalho KI,Maeda S,Marti L,Yamashita J,Haslett PA,Kallas EG
25-0259 was used in Flow cytometry/Cell sorting to explore several aspects of cellular immunity in patients co-infected with human immunodeficiency virus-1 and Mycobacterium leprae.
Sun Jun 01 00:00:00 EDT 2008