Product References
Enhancement of the Soluble Form of OX40 and OX40L Costimulatory Molecules but Reduction of the Membrane Form in Type 1 Diabetes (T1D).
Journal of immunology research
An J,Ding S,Li S,Sun L,Chang X,Huang Z,Zhou B,Fang C,Liu C,Zhang X
12-5905 was used in Flow cytometry/Cell sorting to analyze the expression of membrane OX40 and OX40L (mOX40 and mOX40L) and levels of soluble OX40 and OX40L (sOX40 and sOX40L) in T1D patients to determine their clinical significance.
Fri Jan 24 00:00:00 UTC 2020
Selective HDAC6 inhibitors improve anti-PD-1 immune checkpoint blockade therapy by decreasing the anti-inflammatory phenotype of macrophages and down-regulation of immunosuppressive proteins in tumor cells.
Scientific reports
Knox T,Sahakian E,Banik D,Hadley M,Palmer E,Noonepalle S,Kim J,Powers J,Gracia-Hernandez M,Oliveira V,Cheng F,Chen J,Barinka C,Pinilla-Ibarz J,Lee NH,Kozikowski A,Villagra A
Published figure using CD252 (OX40 Ligand) monoclonal antibody (Product # 12-5905-82) in Flow Cytometry
Tue Apr 16 00:00:00 UTC 2019
Tissue-Restricted Adaptive Type 2 Immunity Is Orchestrated by Expression of the Costimulatory Molecule OX40L on Group 2 Innate Lymphoid Cells.
Immunity
Halim TYF,Rana BMJ,Walker JA,Kerscher B,Knolle MD,Jolin HE,Serrao EM,Haim-Vilmovsky L,Teichmann SA,Rodewald HR,Botto M,Vyse TJ,Fallon PG,Li Z,Withers DR,McKenzie ANJ
12-5905-82 was used in Flow cytometry/Cell sorting to show that the increased expression of OX40L in response to IL-33 acts as a licensing signal in the orchestration of tissue-specific adaptive type 2 immunity, without which this response fails to establish.
Tue Jun 19 00:00:00 UTC 2018
An OX40/OX40L interaction directs successful immunity to hepatitis B virus.
Science translational medicine
Publicover J,Gaggar A,Jespersen JM,Halac U,Johnson AJ,Goodsell A,Avanesyan L,Nishimura SL,Holdorf M,Mansfield KG,Judge JB,Koshti A,Croft M,Wakil AE,Rosenthal P,Pai E,Cooper S,Baron JL
12-5905 was used in Flow cytometry/Cell sorting to study the mechanistic action of the immune pathways and cells necessary for HBV immunity and identify potential therapeutic targets for resolving chronic HBV infection.
Wed Mar 21 00:00:00 UTC 2018
Nitric oxide dependent signaling via cyclic GMP in dendritic cells regulates migration and T-cell polarization.
Scientific reports
Gnipp S,Mergia E,Puschkarow M,Bufe A,Koesling D,Peters M
12-5905 was used in Flow cytometry/Cell sorting to show that activity of NO-GC1 in DCs is important for the magnitude and bias of the Th response in allergic airway disease most likely by counteracting intracellular cAMP.
Fri Jul 20 00:00:00 UTC 2018
Control of Treg cell homeostasis and immune equilibrium by Lkb1 in dendritic cells.
Nature communications
Chen S,Fang L,Guo W,Zhou Y,Yu G,Li W,Dong K,Liu J,Luo Y,Wang B,Li Z,Zhao C,Sun Z,Shen Y,Leng Q,Zhou D,Han Z,Huang H,Ren H,Xu G,Feng X
12-5905 was used in Flow cytometry/Cell sorting to identify Lkb1 as a regulatory switch in dendritic cells for controlling regulatory T cell homeostasis, immune response and tolerance.
Thu Dec 13 00:00:00 UTC 2018
Dectin-1-activated dendritic cells trigger potent antitumour immunity through the induction of Th9 cells.
Nature communications
Zhao Y,Chu X,Chen J,Wang Y,Gao S,Jiang Y,Zhu X,Tan G,Zhao W,Yi H,Xu H,Ma X,Lu Y,Yi Q,Wang S
12-5905 was used in Flow cytometry/Cell sorting to identify dectin-1-activated dendritic cells as a powerful inducer of Th9 cells and antitumour immunity.
Fri Aug 05 00:00:00 UTC 2016
Dectin-1-activated dendritic cells trigger potent antitumour immunity through the induction of Th9 cells.
Nature communications
Zhao Y,Chu X,Chen J,Wang Y,Gao S,Jiang Y,Zhu X,Tan G,Zhao W,Yi H,Xu H,Ma X,Lu Y,Yi Q,Wang S
12-5905 was used in Flow cytometry/Cell sorting to identify dectin-1-activated dendritic cells as a powerful inducer of Th9 cells and antitumour immunity.
Fri Aug 05 00:00:00 UTC 2016
Critical Role of Transcription Factor PU.1 in the Function of the OX40L/TNFSF4 Promoter in Dendritic Cells.
Scientific reports
Yashiro T,Hara M,Ogawa H,Okumura K,Nishiyama C
12590582 was used in flow cytometry to study the role of PU.1 in the expression of OX40L in dendritic cells
Thu Oct 06 00:00:00 UTC 2016
The transcription factor STAT5 is critical in dendritic cells for the development of TH2 but not TH1 responses.
Nature immunology
Bell BD,Kitajima M,Larson RP,Stoklasek TA,Dang K,Sakamoto K,Wagner KU,Kaplan DH,Reizis B,Hennighausen L,Ziegler SF
12-5905 was used in Flow cytometry/Cell sorting to investigate how type-2 immunity is affected by STAT5 transcription factor in dendritic cells.
Mon Apr 01 00:00:00 UTC 2013
Neutralization of TSLP inhibits airway remodeling in a murine model of allergic asthma induced by chronic exposure to house dust mite.
PloS one
Chen ZG,Zhang TT,Li HT,Chen FH,Zou XL,Ji JZ,Chen H
12-5905 was used in Flow cytometry/Cell sorting to suggest that thymic stromal lymphopoietin influences the initiation and persistence of airway inflammation and remodeling during chronic allergic asthma.
Tue Aug 20 00:00:00 UTC 2013
OX40 ligand and programmed cell death 1 ligand 2 expression on inflammatory dendritic cells regulates CD4 T cell cytokine production in the lung during viral disease.
Journal of immunology (Baltimore, Md. : 1950)
Wythe SE,Dodd JS,Openshaw PJ,Schwarze J
12-5905 was used in Flow cytometry/Cell sorting to study the regulation of CD4 T cell cytokine release during viral lung disease, by OX40 and PD-L2 expression on inflammatory dendritic cells.
Wed Feb 15 00:00:00 UTC 2012
Dibutyl phthalate-induced thymic stromal lymphopoietin is required for Th2 contact hypersensitivity responses.
Journal of immunology (Baltimore, Md. : 1950)
Larson RP,Zimmerli SC,Comeau MR,Itano A,Omori M,Iseki M,Hauser C,Ziegler SF
12-5905 was used in Flow cytometry/Cell sorting to demonstrate a role for TSLP in a Th2 model of contact hypersensitivity in mice.
Mon Mar 15 00:00:00 UTC 2010
Cell-cell cooperation at the T helper cell/mast cell immunological synapse.
Blood
Gaudenzio N,Espagnolle N,Mars LT,Liblau R,Valitutti S,Espinosa E
12-5905 was used in Flow cytometry/Cell sorting to investigate whether cognate interactions between mast cells and class II-restricted CD4(+) T cells occur.
Thu Dec 03 00:00:00 UTC 2009