Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) EDA, rmNeogenin, recombinant human TWEAK R, rmOPG, or rmTNF R1 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
TWEAK is a member of the TNF family and was first described as a weak inducer of apoptosis in demonstrate that this relatively small molecule is a fully transformed cell lines. Recently, a receptor for TWEAK was isolated by expression cloning from a HUVEC cell cDNA library. TweakR is a small protein belonging to TNF receptor super family. TweakR is capable of intiating a proliferative signal by binding to TRAF1, 2, and 3. TweakR mRNA is upregulated during tissue regeneration in the aorta and liver. It may also play a role in vascular cell migration and angiogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD266; FGF-inducible 14; fibroblast growth factor regulated protein 2; Fibroblast growth factor-inducible immediate-early response protein 14; Fibroblast growth factor-regulated protein 2; Tumor necrosis factor receptor superfamily member 12A; Tweak-receptor; TweakR; type I transmembrane protein Fn14
Gene Aliases: AI255180; C87282; Fgfrp2; Fn14; HPIP; Tnfrsf12a; TWEAK-R; TweakR
UniProt ID: (Mouse) Q9CR75
Entrez Gene ID: (Mouse) 27279
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support