Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is human spleen tissue lysate.
PA5-20013 can be used with blocking peptide PEP-0132.
Tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), also known as BAFFR, is a protein in humans is encoded by the TNFRSF13C gene. The BAFFR gene is mapped to chromosome 22q13. 1-q13. 31. It has got 184 amino acid transmembrane protein which is 56% identical to the mouse protein. B cell-activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. BAFF plays a crucial role in B cell development and can function through receptors other than BCMA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B cell-activating factor receptor; B-cell maturation defect; B-cell maturation defect 1; B-cell-activating factor receptor; BAFF receptor; BAFF-R; BLyS receptor 3; CD268; Tumor necrosis factor receptor superfamily member 13C; tumor necrosis factor receptor superfamily, member 13C
Gene Aliases: 2010006P15Rik; BAFF-R; BAFFR; Bcmd; Bcmd-1; Bcmd1; BR3; BROMIX; CD268; CVID4; Lvis22; prolixin; RGD1560810; TNFRSF13C
UniProt ID: (Human) Q96RJ3, (Mouse) Q9D8D0
Entrez Gene ID: (Human) 115650, (Rat) 500910, (Mouse) 72049
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support