Save $650 & get a lab-themed hidden object puzzle

Save and claim

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • PD-L1 (CD274) Antibodies

            Invitrogen

            CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH1), PerCP-eFluor™ 710, eBioscience™

            87 Published Figures
            60 References
            View all (247) PD-L1 (CD274) antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 46-5983-42
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            379.10
            Special offer
            Online exclusive
            446.00
            Save 66.90 (15%)
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH1), PerCP-eFluor™ 710, eBioscience™

            • Antibody Testing Data (1)
            • Published Figures (87)
            Application
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 88

            CD274 (PD-L1, B7-H1) Antibody (46-5983-42) in Flow

            Staining of unstimulated (left) or PHA-stimulated (right) normal human peripheral blood cells with Mouse IgG1 K Isotype Control PerCP-eFluor® 710 (Product # 46-4714-82) (blue histogram) or Anti-Human CD274 (B7-H1) PerCP-eFluor® 710 (purple histogram). Total viable cells were used for analysis.
            View Product

            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
            CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)

            Product Details

            46-5983-42

            Applications
            Tested Dilution
            Publications

            Immunohistochemistry (Frozen) (IHC (F))

            -
            View 2 publications 2 publications

            Flow Cytometry (Flow)

            5 µL (1 µg)/test
            View 56 publications 56 publications

            ChIP assay (ChIP)

            -
            View 1 publication 1 publication

            Functional Assay (Functional)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human

            Published species

            Human, Mouse

            Host/Isotype

            Mouse / IgG1, kappa

            Recommended Isotype Control

            Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), PerCP-eFluor™ 710, eBioscience™

            Class

            Monoclonal

            Type

            Antibody

            Clone

            MIH1

            Conjugate

            PerCP-eFluor™ 710
            • Unconjugated
            • Alexa Fluor 488 
            • Alexa Fluor 700 
            • APC 
            • APC-eFluor 780
            • Biotin 
            • Brilliant Violet 786
            • eFluor 450
            • Functional Grade
            • NovaFluor Red 685
            • NovaFluor Red 700
            • NovaFluor Yellow 700
            • PE 
            • PE-Cyanine5
            • PE-Cyanine7
            • PE-eFluor 610
            • Request custom conjugation

            Excitation/Emission Max

            482/708 nm View spectra spectra

            Form

            Liquid

            Concentration

            5 µL/Test

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2, with BSA

            Contains

            0.09% sodium azide

            Storage conditions

            4°C, store in dark, DO NOT FREEZE!

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_11041815

            Product Specific Information

            Description: The MIH1 monoclonal antibody reacts with human B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40 kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH1 is reported to be a blocking antibody.

            Applications Reported: This MIH1 antibody has been reported for use in flow cytometric analysis.

            Applications Tested: This MIH1 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated human peripheral blood cells. This can be used at 5 µL (1 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

            PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative.

            Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols.

            Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            How to build a flow cytometry experiment with Invitrogen Flow Cytometry Panel Builder
            The Invitrogen Flow Cytometry Panel Builder webtool helps you chose antibody clones, fluorophores, fluorescent proteins, and reagents to design your experiment.
            Video Player is loading.
            Current Time 0:00
            /
            Duration 8:21
            Loaded: 0.00%
            0:00
            Stream Type LIVE
            Remaining Time -8:21
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected
            • en (Main), selected

            This is a modal window.

            Beginning of dialog window. Escape will cancel and close the window.

            Text
            Text Background
            Caption Area Background
            Font Size
            Text Edge Style
            Font Family

            End of dialog window.

            This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

            How to use the Panel Builder

            Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

            Bioinformatics

            Protein Aliases: B7 homolog 1; B7-H1; CD274; CD274 antigen; PD-L1; PDCD1 ligand 1; pdl-1; Programmed cell death 1 ligand 1; sCD274; soluble CD274; soluble PD L1; sPD L1

            View more View less

            Gene Aliases: B7-H; B7H1; CD274; PD-L1; PDCD1L1; PDCD1LG1; PDL1

            View more View less

            UniProt ID: (Human) Q9NZQ7

            View more View less

            Entrez Gene ID: (Human) 29126

            View more View less

            Function(s)

            protein binding immunoglobulin receptor superfamily

            Process(es)

            immune response signal transduction cell surface receptor signaling pathway positive regulation of cell migration T cell costimulation negative regulation of interferon-gamma production negative regulation of interleukin-10 production response to cytokine positive regulation of T cell proliferation regulation of activated T cell proliferation negative regulation of activated T cell proliferation regulation of T cell apoptotic process toxin transport negative regulation of tumor necrosis factor superfamily cytokine production positive regulation of interleukin-10 secretion
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-pxmrt:80/100.66.79.171:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0