Product References
Hedgehog costimulation during ischemia-reperfusion injury potentiates cytokine and homing responses of CD4+ T cells.
Frontiers in immunology
Wang S,Song G,Barkestani MN,Tobiasova Z,Wang Q,Jiang Q,Lopez R,Adelekan-Kamara Y,Fan M,Pober JS,Tellides G,Jane-Wit D
12-2799-42 was used in Flow Cytometry to develop a humanized mouse model to resolve functional and migratory heterogeneity within the PtchHi population.
Fri Nov 03 00:00:00 UTC 2023
Hedgehog-induced ZFYVE21 promotes chronic vascular inflammation by activating NLRP3 inflammasomes in T cells.
Science signaling
Jiang B,Wang S,Song G,Jiang Q,Fan M,Fang C,Li X,Soh CL,Manes TD,Cheru N,Qin L,Ren P,Jortner B,Wang Q,Quaranta E,Yoo P,Geirsson A,Davis RP,Tellides G,Pober JS,Jane-Wit D
12-2799-42 was used in Flow cytometry/Cell sorting to identify a T cell-autonomous role for ZFYVE21 in promoting chronic vascular inflammation associated with allograft vasculopathy.
Tue Mar 21 00:00:00 UTC 2023
Glucose metabolism controls human γδ T-cell-mediated tumor immunosurveillance in diabetes.
Cellular & molecular immunology
Mu X,Xiang Z,Xu Y,He J,Lu J,Chen Y,Wang X,Tu CR,Zhang Y,Zhang W,Yin Z,Leung WH,Lau YL,Liu Y,Tu W
12-2799-42 was used in Flow Cytometry to suggest that the impaired antitumor activity of Vγ9Vδ2 T cells induced by dysregulated glucose metabolism may contribute to the increased cancer risk in T2DM patients and that metabolic reprogramming by targeting the AMPK pathway with metformin may improve tumor immunosurveillance.
Mon Aug 01 00:00:00 UTC 2022
Sex difference in the expression of PD-1 of non-small cell lung cancer.
Frontiers in immunology
Gu Y,Tang YY,Wan JX,Zou JY,Lu CG,Zhu HS,Sheng SY,Wang YF,Liu HC,Yang J,Hong H
12-2799-42 was used in Flow cytometry/Cell sorting to examine sex differences in serum sPD-1, mPD-1 expression on T cells, and sex hormone levels in non-small cell lung cancer (NSCLC) patients.
Tue Nov 08 00:00:00 UTC 2022
Immunogenic Chemotherapy Enhances Recruitment of CAR-T Cells to Lung Tumors and Improves Antitumor Efficacy when Combined with Checkpoint Blockade.
Cancer cell
Srivastava S,Furlan SN,Jaeger-Ruckstuhl CA,Sarvothama M,Berger C,Smythe KS,Garrison SM,Specht JM,Lee SM,Amezquita RA,Voillet V,Muhunthan V,Yechan-Gunja S,Pillai SPS,Rader C,Houghton AM,Pierce RH,Gottardo R,Maloney DG,Riddell SR
12-2799-42 was used in Flow Cytometry to test strategies for enhancing efficacy of CAR-T cells targeting the tumor-associated antigen receptor tyrosine kinase-like orphan receptor 1 (ROR1) infiltrating tumors.
Mon Feb 08 00:00:00 UTC 2021
CAR T cells targeting tumor-associated exons of glypican 2 regress neuroblastoma in mice.
Cell reports. Medicine
Li N,Torres MB,Spetz MR,Wang R,Peng L,Tian M,Dower CM,Nguyen R,Sun M,Tai CH,de Val N,Cachau R,Wu X,Hewitt SM,Kaplan RN,Khan J,St Croix B,Thiele CJ,Ho M
12-2799-42 was used in Flow cytometry/Cell sorting to investigate if RNA-seq data can be exploited to help identify tumor-associated exons that can be targeted by CAR T cell therapies.
Tue Jun 15 00:00:00 UTC 2021
Harnessing Natural Killer Immunity in Metastatic SCLC.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer
Best SA,Hess JB,Souza-Fonseca-Guimaraes F,Cursons J,Kersbergen A,Dong X,Rautela J,Hyslop SR,Ritchie ME,Davis MJ,Leong TL,Irving L,Steinfort D,Huntington ND,Sutherland KD
12-2799 was used in Flow cytometry/Cell sorting to investigate the immunophenotype of human SCLC tumours using multiple forms of analysis including flow cytometry using human samples.
Tue Sep 01 00:00:00 UTC 2020
Enhanced CAR-T activity against established tumors by polarizing human T cells to secrete interleukin-9.
Nature communications
Liu L,Bi E,Ma X,Xiong W,Qian J,Ye L,Su P,Wang Q,Xiao L,Yang M,Lu Y,Yi Q
12-2799 was used in Flow cytometry/Cell sorting to show that, through a Th9-culture condition, polarized and expanded human CAR-T cells have an enhanced antitumor activity.
Thu Nov 19 00:00:00 UTC 2020
Circulating Exosomes Inhibit B Cell Proliferation and Activity.
Cancers
Schroeder JC,Puntigam L,Hofmann L,Jeske SS,Beccard IJ,Doescher J,Laban S,Hoffmann TK,Brunner C,Theodoraki MN,Schuler PJ
12-2799 was used in Flow cytometry/Cell sorting to isolate and analyse peripheral B cells by flow cytometry to understand the effects of exosomes on B cells.
Wed Jul 29 00:00:00 UTC 2020
A facile technology for the high-throughput sequencing of the paired VH:VL and TCRβ:TCRα repertoires.
Science advances
Tanno H,McDaniel JR,Stevens CA,Voss WN,Li J,Durrett R,Lee J,Gollihar J,Tanno Y,Delidakis G,Pothukuchy A,Ellefson JW,Goronzy JJ,Maynard JA,Ellington AD,Ippolito GC,Georgiou G
12-2799 was used in Flow cytometry/Cell sorting to determine the VH:VL or TCRb:a repertoire with high accuracy and throughput.
Wed Apr 01 00:00:00 UTC 2020
Th2 Biased Immunity With Altered B Cell Profiles in Circulation of Patients With Sporotrichosis Caused by Sporothrix globosa.
Frontiers in immunology
Zu J,Yao L,Song Y,Cui Y,Guan M,Chen R,Zhen Y,Li S
12-2799 was used in Flow cytometry/Cell sorting to shed light on the potential involvement of peripheral T and B cell immunity against this mycotic infection.
Mon May 31 00:00:00 UTC 2021
Circulating exosomes measure responses to therapy in head and neck cancer patients treated with cetuximab, ipilimumab, and IMRT.
Oncoimmunology
Theodoraki MN,Yerneni S,Gooding WE,Ohr J,Clump DA,Bauman JE,Ferris RL,Whiteside TL
12-2799-42 was used in Flow cytometry/Cell sorting to support the potential role of exosomes as a non-invasive tumor and immune cell biomarkers in cancer.
Tue Jan 12 00:00:00 UTC 2021
Separation of plasma-derived exosomes into CD3(+) and CD3(-) fractions allows for association of immune cell and tumour cell markers with disease activity in HNSCC patients.
Clinical and experimental immunology
Theodoraki MN,Hoffmann TK,Whiteside TL
12-2799 was used in Flow cytometry/Cell sorting to investigate whether tumour- or T cell-derived exosomes in head and neck squamous cell carcinoma patients' plasma are immunosuppressive and impact upon disease activity.
Fri Jun 01 00:00:00 UTC 2018
Phenotypic and Functional Profiles of Antigen-Specific CD4+ and CD8+ T Cells Associated With Infection Control in Patients With Cutaneous Leishmaniasis.
Frontiers in cellular and infection microbiology
Egui A,Ledesma D,Pérez-Antón E,Montoya A,Gómez I,Robledo SM,Infante JJ,Vélez ID,López MC,Thomas MC
12-2799 was used in Flow cytometry/Cell sorting to suggest a possible exhaustion process of CD8+ T cells associated with the evolution of Leishmania infection.
Wed Sep 18 00:00:00 UTC 2019
YY1 Upregulates Checkpoint Receptors and Downregulates Type I Cytokines in Exhausted, Chronically Stimulated Human T Cells.
iScience
Balkhi MY,Wittmann G,Xiong F,Junghans RP
12-2799 was used in Flow cytometry/Cell sorting to elucidate a Yin Yang-1-centred mechanism for diverse cellular components correlated with exhaustion.
Fri Apr 27 00:00:00 UTC 2018
Immune evasion mediated by PD-L1 on glioblastoma-derived extracellular vesicles.
Science advances
Ricklefs FL,Alayo Q,Krenzlin H,Mahmoud AB,Speranza MC,Nakashima H,Hayes JL,Lee K,Balaj L,Passaro C,Rooj AK,Krasemann S,Carter BS,Chen CC,Steed T,Treiber J,Rodig S,Yang K,Nakano I,Lee H,Weissleder R,Breakefield XO,Godlewski J,Westphal M,Lamszus K,Freeman GJ,Bronisz A,Lawler SE,Chiocca EA
12-2799 was used in Flow cytometry/Cell sorting to investigate whether glioblastoma extracellular vesicles are important mediators of immunosuppression and whether programmed death ligand-1 could play a role.
Thu Mar 01 00:00:00 UTC 2018
The influence of chemotherapy on adenosine-producing B cells in patients with head and neck squamous cell carcinoma.
Oncotarget
Ziebart A,Huber U,Jeske S,Laban S,Doescher J,Hoffmann TK,Brunner C,Jackson EK,Schuler PJ
12-2799 was used in Flow cytometry/Cell sorting to study the influence of chemotherapy on the function of regulatory B cells.
Fri Jan 19 00:00:00 UTC 2018
Overcoming Target Driven Fratricide for T Cell Therapy.
Frontiers in immunology
Breman E,Demoulin B,Agaugué S,Mauën S,Michaux A,Springuel L,Houssa J,Huberty F,Jacques-Hespel C,Marchand C,Marijsse J,Nguyen T,Ramelot N,Violle B,Daro D,De Waele P,Gilham DE,Steenwinckel V
12-2799 was used in Flow cytometry/Cell sorting to demonstrate that target-driven chimeric antigen receptor T cell fratricide can be overcome through targeted inhibition of NKG2D expression or inhibition of enzyme function.
Mon Oct 07 00:00:00 UTC 2019
Cytomegalovirus infection in HIV-infected and uninfected individuals is characterized by circulating regulatory T cells of unconstrained antigenic specificity.
PloS one
Tovar-Salazar A,Weinberg A
12-2799 was used in Flow cytometry/Cell sorting to indicate that CMV-seropositive individuals have circulating Tregs that inhibit cell-mediated immune responses to CMV and other antigens and may be contribute to an immune-suppressive effect of CMV infection.
Fri Oct 06 00:00:00 UTC 2017
Tumor-derived exosomes regulate expression of immune function-related genes in human T cell subsets.
Scientific reports
Muller L,Mitsuhashi M,Simms P,Gooding WE,Whiteside TL
12-2799 was used in Flow cytometry/Cell sorting to study the regulation of immune function-related genes in human T cell subsets through tumor-derived exosomes.
Thu Feb 04 00:00:00 UTC 2016
Inhibitory Receptor Expression on CD8+ T Cells Is Linked to Functional Responses against Trypanosoma cruzi Antigens in Chronic Chagasic Patients.
Journal of immunology (Baltimore, Md. : 1950)
Lasso P,Mateus J,Pavía P,Rosas F,Roa N,Thomas MC,López MC,González JM,Puerta CJ,Cuéllar A
12-2799 was used in Flow cytometry/Cell sorting to suggest that during Chagas disease, CD8(+) T cells undergo gradual loss of function, impaired cytokine production and increased inhibitory receptor coexpression.
Thu Oct 15 00:00:00 UTC 2015
Lipopolysaccharide Increases Immune Activation and Alters T Cell Homeostasis in SHIVB'WHU Chronically Infected Chinese Rhesus Macaque.
Journal of immunology research
Zhang GH,Wu RD,Zheng HY,Zhang XL,Zhang MX,Tian RR,Liu GM,Pang W,Zheng YT
12-2799 was used in Flow cytometry/Cell sorting to investigate the mechanisms behind lipopolysaccharide disruption of immune responses in Chinese rhesus macaques.
Wed Sep 07 00:00:00 UTC 2016
Circulating CXCR5⁺CD4⁺ T Follicular-Like Helper Cell and Memory B Cell Responses to Human Papillomavirus Vaccines.
PloS one
Matsui K,Adelsberger JW,Kemp TJ,Baseler MW,Ledgerwood JE,Pinto LA
12-2799 was used in Flow cytometry/Cell sorting to identify different subsets of CXCR5+CD4+ Tfh-like cells in response to highly immunogenic and efficacious vaccines for HPV.
Tue May 24 00:00:00 UTC 2016
Exosomes isolated from plasma of glioma patients enrolled in a vaccination trial reflect antitumor immune activity and might predict survival.
Oncoimmunology
Muller L,Muller-Haegele S,Mitsuhashi M,Gooding W,Okada H,Whiteside TL
12-2799 was used in Flow cytometry/Cell sorting to show exosomes from the plasma of glioma patients from a vaccination trial reflect antitumour immune activity which may predict survival.
Mon Jun 01 00:00:00 UTC 2015
Rapid Proliferation and Differentiation of a Subset of Circulating IgM Memory B Cells to a CpG/Cytokine Stimulus In Vitro.
PloS one
Vásquez C,Franco MA,Angel J
12-2799 was used in Flow cytometry/Cell sorting to compare the phenotype and in vitro functional response of various immune cells in response to CpG and a cytokine cocktail.
Thu Jun 02 00:00:00 UTC 2016
Failure to upregulate cell surface PD-1 is associated with dysregulated stimulation of T cells by TGN1412-like CD28 superagonist.
mAbs
Thaventhiran T,Alhumeed N,Yeang HX,Sethu S,Downey JS,Alghanem AF,Olayanju A,Smith EL,Cross MJ,Webb SD,Williams DP,Bristow A,Ball C,Stebbings R,Sathish JG
12-2799 was used in Flow cytometry/Cell sorting to understand the underlying molecular mechanisms of T-cell activation in response to TGN1412-like CD28 superagonist, showing that failure to upregulate the inhibitory PD-1 cell surface receptor is part of this mechanism.
Mon Aug 24 00:00:00 UTC 2015
Changes in immune cell populations in the periphery and liver of GBV-B-infected and convalescent tamarins (Saguinus labiatus).
Virus research
Hood SP,Mee ET,Perkins H,Bowen O,Dale JM,Almond NM,Karayiannis P,Bright H,Berry NJ,Rose NJ
12-2799 was used in Flow cytometry/Cell sorting to characterise changes in lymphocyte populations in tamarins with an acute GBV-B infection.
Wed Jan 22 00:00:00 UTC 2014
Comparison of influenza and SIV specific CD8 T cell responses in macaques.
PloS one
Jegaskanda S,Reece JC,De Rose R,Stambas J,Sullivan L,Brooks AG,Kent SJ,Sexton A
12-2799 was used in Flow cytometry/Cell sorting to suggest that strategies to maintain a more functional CD8(+) T-cell response, profile may assist in controlling HIV disease.
Mon Jul 09 00:00:00 UTC 2012
Increased frequency of regulatory T cells accompanies increased immune activation in rectal mucosae of HIV-positive noncontrollers.
Journal of virology
Shaw JM,Hunt PW,Critchfield JW,McConnell DH,Garcia JC,Pollard RB,Somsouk M,Deeks SG,Shacklett BL
12-2799 was used in Flow cytometry/Cell sorting to investigate the role of Tregs in persistent HIV infection, showing that an increased frequency of Tregs accompanies increased immune activation in HIV-positive noncontrollers.
Tue Nov 01 00:00:00 UTC 2011
Phagocytosis, a potential mechanism for myeloid-derived suppressor cell regulation of CD8+ T cell function mediated through programmed cell death-1 and programmed cell death-1 ligand interaction.
Journal of immunology (Baltimore, Md. : 1950)
Kim YJ,Park SJ,Broxmeyer HE
12-2799 was used in Flow cytometry/Cell sorting to identify a potential mechanism by which IFN-γ-producing CD8(+) T cells are tolerised after type 1 immune responses to chronic virus or tumour.
Thu Sep 01 00:00:00 UTC 2011