Product References
Aging affects responsiveness of peripheral blood mononuclear cells to immunosuppression of periodontal ligament stem cells.
The Journal of international medical research
Li X,Zhang B,Wang H,Zhao X,Zhang Z,Ding G,Wei F
Published figure using CD283 (TLR3) monoclonal antibody (Product # 12-9039-82) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2020
Aging affects responsiveness of peripheral blood mononuclear cells to immunosuppression of periodontal ligament stem cells.
The Journal of international medical research
Li X,Zhang B,Wang H,Zhao X,Zhang Z,Ding G,Wei F
Published figure using CD283 (TLR3) monoclonal antibody (Product # 12-9039-82) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2020
The expression profile of virus-recognizing toll-like receptors in natural killer cells of Cypriot multiple sclerosis patients.
BMC research notes
Deeba E,Lambrianides A,Pantzaris M,Krashias G,Christodoulou C
12-9039-82 was used in Flow Cytometry to study toll-like receptor (TLR) expression in the natural killer cells of patients with multiple sclerosis.
Tue Sep 29 00:00:00 UTC 2020
Hepatitis C virus infection induces autocrine interferon signaling by human liver endothelial cells and release of exosomes, which inhibits viral replication.
Gastroenterology
Giugliano S,Kriss M,Golden-Mason L,Dobrinskikh E,Stone AE,Soto-Gutierrez A,Mitchell A,Khetani SR,Yamane D,Stoddard M,Li H,Shaw GM,Edwards MG,Lemon SM,Gale M,Shah VH,Rosen HR
12-9039 was used in Flow cytometry/Cell sorting to demonstrate that cultured human liver sinusoidal endothelial cells produce factors that mediate immunity against hepatitis C virus during infection.
Sun Feb 01 00:00:00 UTC 2015
Hepatitis C Virus Sensing by Human Trophoblasts Induces Innate Immune Responses and Recruitment of Maternal NK Cells: Potential Implications for Limiting Vertical Transmission.
Journal of immunology (Baltimore, Md. : 1950)
Giugliano S,Petroff MG,Warren BD,Jasti S,Linscheid C,Ward A,Kramer A,Dobrinskikh E,Sheiko MA,Gale M,Golden-Mason L,Winn VD,Rosen HR
12-9039 was used in Flow cytometry/Cell sorting to demonstrate that HCV-RNA sensing by human trophoblasts induces recruitment and activation of innate immune cells.
Thu Oct 15 00:00:00 UTC 2015
A potential mode of action for Anakinra in patients with arthrofibrosis following total knee arthroplasty.
Scientific reports
Dixon D,Coates J,del Carpio Pons A,Horabin J,Walker A,Abdul N,Kalson NS,Brewster NT,Weir DJ,Deehan DJ,Mann DA,Borthwick LA
12-9039 was used in Flow cytometry/Cell sorting to propose a potential mechanism for the reported beneficial impact of Anakinra on patients with arthrofibrosis.
Tue Nov 10 00:00:00 UTC 2015
A potential mode of action for Anakinra in patients with arthrofibrosis following total knee arthroplasty.
Scientific reports
Dixon D,Coates J,del Carpio Pons A,Horabin J,Walker A,Abdul N,Kalson NS,Brewster NT,Weir DJ,Deehan DJ,Mann DA,Borthwick LA
12-9039 was used in Flow cytometry/Cell sorting to propose a potential mechanism for the reported beneficial impact of Anakinra on patients with arthrofibrosis.
Tue Nov 10 00:00:00 UTC 2015
Syndrome of selective IgM deficiency with severe T cell deficiency associated with disseminated cutaneous mycobacterium avium intracellulaire infection.
American journal of clinical and experimental immunology
Gharib A,Louis AG,Agrawal S,Gupta S
12-9039 was used in Flow cytometry/Cell sorting to analyse the selective IgM deficiency and T cell lymphocytopenia associated with disseminated cutaneous Mycobacterium avium intracellulaire infection.
Mon Nov 09 00:00:00 UTC 2015
IL-29 enhances Toll-like receptor-mediated IL-6 and IL-8 production by the synovial fibroblasts from rheumatoid arthritis patients.
Arthritis research & therapy
Xu L,Feng X,Tan W,Gu W,Guo D,Zhang M,Wang F
12-9039 was used in Flow cytometry/Cell sorting to investigate the effect of IL-29 on cytokine production in synovial fibroblasts in arthritis patients.
Tue Oct 29 00:00:00 UTC 2013
Identification of lineage relationships and novel markers of blood and skin human dendritic cells.
Journal of immunology (Baltimore, Md. : 1950)
Harman AN,Bye CR,Nasr N,Sandgren KJ,Kim M,Mercier SK,Botting RA,Lewin SR,Cunningham AL,Cameron PU
12-9039 was used in Flow cytometry/Cell sorting to study the lineage relationships between blood and tissue dendritic cells, providing novel markers of blood and skin human dendritic cells.
Tue Jan 01 00:00:00 UTC 2013
Bone marrow mononuclear cells up-regulate toll-like receptor expression and produce inflammatory mediators in response to cigarette smoke extract.
PloS one
Zhou J,Eksioglu EA,Fortenbery NR,Chen X,Wang H,Epling-Burnette PK,Djeu JY,Wei S
12-9039 was used in Flow cytometry/Cell sorting to elucidate the effects of cigarette smoke on human bone marrow haematopoiesis and characterise the inflammatory response.
Thu Dec 01 00:00:00 UTC 2011
Porcine small intestinal epithelial cell line (IPEC-J2) of rotavirus infection as a new model for the study of innate immune responses to rotaviruses and probiotics.
Viral immunology
Liu F,Li G,Wen K,Bui T,Cao D,Zhang Y,Yuan L
12-9039 was used in Flow cytometry/Cell sorting to establish a novel in-vitro model of rotavirus infection and probiotic treatment.
Thu Apr 01 00:00:00 UTC 2010
Secretion of the human Toll-like receptor 3 ectodomain is affected by single nucleotide polymorphisms and regulated by Unc93b1.
The Journal of biological chemistry
Qi R,Hoose S,Schreiter J,Sawant KV,Lamb R,Ranjith-Kumar CT,Mills J,San Mateo L,Jordan JL,Kao CC
12-9039 was used in Flow cytometry/Cell sorting to show that a siRNA knockdown of Unc93b1 can reduce TLR3 abundance on the cell surface without affecting total TLR3 accumulation.
Fri Nov 19 00:00:00 UTC 2010
Small interfering RNA-induced TLR3 activation inhibits blood and lymphatic vessel growth.
Proceedings of the National Academy of Sciences of the United States of America
Cho WG,Albuquerque RJ,Kleinman ME,Tarallo V,Greco A,Nozaki M,Green MG,Baffi JZ,Ambati BK,De Falco M,Alexander JS,Brunetti A,De Falco S,Ambati J
12-9039 was used in Flow cytometry/Cell sorting to introduce TLR3 activation as a method of jointly suppressing blood and lymphatic neovascularization.
Tue Apr 28 00:00:00 UTC 2009
Toll-like receptor and innate cytokine responses induced by lactobacilli colonization and human rotavirus infection in gnotobiotic pigs.
Veterinary immunology and immunopathology
Wen K,Azevedo MS,Gonzalez A,Zhang W,Saif LJ,Li G,Yousef A,Yuan L
12-9039 was used in Flow cytometry/Cell sorting to evaluate TLR2-, TLR3- and TLR9-expressing antigen presenting cell responses in the spleens and blood of gnotobiotic pigs.
Sun Feb 15 00:00:00 UTC 2009
Biochemical and functional analyses of the human Toll-like receptor 3 ectodomain.
The Journal of biological chemistry
Ranjith-Kumar CT,Miller W,Xiong J,Russell WK,Lamb R,Santos J,Duffy KE,Cleveland L,Park M,Bhardwaj K,Wu Z,Russell DH,Sarisky RT,Mbow ML,Kao CC
12-9039 was used in Flow cytometry/Cell sorting to determine through mutational analysis the biochemical function of human TLR3 ectodomain.
Fri Mar 09 00:00:00 UTC 2007
Effects of single nucleotide polymorphisms on Toll-like receptor 3 activity and expression in cultured cells.
The Journal of biological chemistry
Ranjith-Kumar CT,Miller W,Sun J,Xiong J,Santos J,Yarbrough I,Lamb RJ,Mills J,Duffy KE,Hoose S,Cunningham M,Holzenburg A,Mbow ML,Sarisky RT,Kao CC
12-9039 was used in Flow cytometry/Cell sorting to examine the effect of single nucleotide polymorphisms on Toll-like receptor 3 signalling and expression.
Fri Jun 15 00:00:00 UTC 2007
Toll-like receptor 2, 3, and 4 expression and function in human airway smooth muscle.
The Journal of allergy and clinical immunology
Sukkar MB,Xie S,Khorasani NM,Kon OM,Stanbridge R,Issa R,Chung KF
12-9039 was used in Flow cytometry/Cell sorting to determine the effects of cytokines, TLR ligands, and corticosteroids on TLR expression in human airway smooth muscle cells.
Fri Sep 01 00:00:00 UTC 2006
Diesel exhaust enhances virus- and poly(I:C)-induced Toll-like receptor 3 expression and signaling in respiratory epithelial cells.
American journal of physiology. Lung cellular and molecular physiology
Ciencewicki J,Brighton L,Wu WD,Madden M,Jaspers I
12-9039 was used in Flow cytometry/Cell sorting to demonstrate that exposure of respiratory epithelial cells to diesel exhaust can increase the expression and function of TLR3.
Thu Jun 01 00:00:00 UTC 2006
Structural and functional analyses of the human Toll-like receptor 3. Role of glycosylation.
The Journal of biological chemistry
Sun J,Duffy KE,Ranjith-Kumar CT,Xiong J,Lamb RJ,Santos J,Masarapu H,Cunningham M,Holzenburg A,Sarisky RT,Mbow ML,Kao C
12-9039 was used in Flow cytometry/Cell sorting to investigate the relationship between toll-like receptor 3 and glycosylation.
Fri Apr 21 00:00:00 UTC 2006
Expansion of toll-like receptor 9-expressing B cells in active systemic lupus erythematosus: implications for the induction and maintenance of the autoimmune process.
Arthritis and rheumatism
Papadimitraki ED,Choulaki C,Koutala E,Bertsias G,Tsatsanis C,Gergianaki I,Raptopoulou A,Kritikos HD,Mamalaki C,Sidiropoulos P,Boumpas DT
12-9039 was used in Flow cytometry/Cell sorting to examine the expression and function of various TLRs in subpopulations of peripheral blood mononuclear cells of patients with systemic lupus erythematosus.
Wed Nov 01 00:00:00 UTC 2006
Toll-like receptor 3 is induced by and mediates antiviral activity against rhinovirus infection of human bronchial epithelial cells.
Journal of virology
Hewson CA,Jardine A,Edwards MR,Laza-Stanca V,Johnston SL
12-9039 was used in Flow cytometry/Cell sorting to demonstrate an important functional requirement for TLR3 in the host response against live virus infection.
Sat Oct 01 00:00:00 UTC 2005
Human endometrial epithelial cells cyclically express Toll-like receptor 3 (TLR3) and exhibit TLR3-dependent responses to dsRNA.
Human immunology
Jorgenson RL,Young SL,Lesmeister MJ,Lyddon TD,Misfeldt ML
Published figure using CD283 (TLR3) monoclonal antibody (Product # 12-9039-82) in Flow Cytometry
Sun May 01 00:00:00 UTC 2005
Human endometrial epithelial cells cyclically express Toll-like receptor 3 (TLR3) and exhibit TLR3-dependent responses to dsRNA.
Human immunology
Jorgenson RL,Young SL,Lesmeister MJ,Lyddon TD,Misfeldt ML
Published figure using CD283 (TLR3) monoclonal antibody (Product # 12-9039-82) in Flow Cytometry
Sun May 01 00:00:00 UTC 2005
17Beta-estradiol suppresses TLR3-induced cytokine and chemokine production in endometrial epithelial cells.
Reproductive biology and endocrinology : RB&E
Lesmeister MJ,Jorgenson RL,Young SL,Misfeldt ML
12-9039 was used in Flow cytometry/Cell sorting to investigate the effects of 17beta-estradiol and progesterone on TLR3 expression and function in endometrial cell lines.
Thu Dec 29 00:00:00 UTC 2005
Activation of airway epithelial cells by toll-like receptor agonists.
American journal of respiratory cell and molecular biology
Sha Q,Truong-Tran AQ,Plitt JR,Beck LA,Schleimer RP
12-9039 was used in Flow cytometry/Cell sorting to investigate the presence and function of Toll-like receptors in the BEAS-2B airway epithelial cell line and primary bronchial epithelial cells.
Wed Sep 01 00:00:00 UTC 2004