Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Description: This three-color cocktail is designed to identify T cells from peripheral blood samples. It can be used alone, or in combination with other antibodies to further characterize T cell populations. The SK3 monoclonal antibody reacts with human CD4, a 59 kDa cell surface receptor expressed by a majority of thymocytes, a subpopulation of mature T helper cells, and in low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). The SK7 monoclonal antibody reacts with human CD3 eplison, a 20 kDa subunit of the CD3/TCR complex. The CD3 complex, including the epsilon, gamma and delta subunits, is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. The SK1 monoclonal antibody reacts with the human CD8a molecule, an approximately 32-34 kDa cell surface receptor expressed either as a heterodimer with the CD8beta chain (CD8alpha beta) or as a homodimer (CD8alphaalpha). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a.
This cocktail contains the following antibodies:
Anti-Human CD4 (SK3) FITC
Anti-Human CD3 (SK7) PerCP-eFluor® 710
Anti-Human CD8a (SK1) APC
Applications Reported: eZfluor Anti-Human CD4 FITC, CD3 PerCP-eFluor® 710, CD8a APC has been reported for use in flow cytometric analysis.
Applications Tested: eZfluor Anti-Human CD4 FITC, CD3 PerCP-eFluor® 710, CD8a APC has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 20 µL per test, which contains 0.06 µg of Anti-Human CD4 FITC, 0.06 µg of Anti-Human CD3 PerCP-eFluor® 710, and 0.25 µg of Anti-Human CD8a APC. A test is defined as the amount of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
CD8 (Cluster of Differentiation 8) is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediate efficient cell-cell interactions within the immune system. The CD8 antigen acts as a co-receptor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20-35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer (NK) cells, subpopulations of peripheral blood null cells, thymocytes and bone marrow cells. The CD8 co-receptor functions as either a homodimer composed of two alpha chains, or as a disulfide-linked heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. The majority of CD8+ T cells express CD8 as a alpha/beta heterodimer. In HIV, the HIV-2 envelope glycoprotein binds CD8 alpha chain (but not the beta chain).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD247; CD247 antigen, zeta subunit; CD3 antigen, delta subunit; CD3 delta; CD3 epsilon; CD3-epsilon; CD3d; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule, delta (CD3-TCR complex); CD3e; CD3e antigen, epsilon polypeptide (TiT3 complex); CD3e molecule, epsilon (CD3-TCR complex); CD3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3Z antigen, zeta polypeptide (TiT3 complex); CD3zeta chain; CD4; CD4 antigen (p55); CD4 antigen p55; CD4 receptor; CD8 antigen, alpha polypeptide (p32); CD8 antigen, beta polypeptide 1 (p37); CD8a; CD8alpha; CD8b; CD8beta; FLJ18683; Leu-2; Leu2 T-lymphocyte antigen; OKT3, delta chain; OKT8 T-cell antigen; T cell co-receptor; T lymphocyte surface glycoprotein beta chain; T-cell antigen Leu2; T-cell antigen receptor complex, epsilon subunit of T3; T-cell antigen receptor complex, gamma subunit of T3; T-cell antigen receptor complex, zeta subunit of CD3; T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; T-cell receptor T3 zeta chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; T-cell surface glycoprotein CD3 zeta chain; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-lymphocyte differentiation antigen T8/Leu-2; T8 T-cell antigen; TCR zeta chain
Gene Aliases: CD247; CD3-DELTA; CD3-GAMMA; CD3-ZETA; CD3D; CD3E; CD3G; CD3H; CD3Q; CD3Z; CD4; CD4mut; CD8; CD8A; CD8B; CD8B1; IMD17; IMD18; IMD19; IMD25; LEU2; LY3; LYT3; MAL; p32; P37; T3D; T3E; T3G; T3Z; TCRE; TCRZ
UniProt ID: (Human) P20963, (Human) P04234, (Human) P07766, (Human) P09693, (Human) P01730, (Human) P01732, (Human) P10966
Entrez Gene ID: (Human) 919, (Human) 915, (Human) 916, (Human) 917, (Human) 920, (Human) 925, (Human) 926
Molecular Function:
defense/immunity protein
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support