Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Positive test controls include: Mouse lung, Rat lung.
Immunogen sequence: MWLPPALLLL SLSGCFSIQG PESVRAPEQG SLTVQCHYKQ GWETYIKWWC RGVRWDTCKI LIETRGSEQG EKSDRVSIKD NQKDRTFTVT MEGLRRDDAD VYWCGIERRG PDLGTQVKVI VDPEGAASTT ASSPTNSNMA VFIGSHKRNH YMLLVFVKVP ILLILVTAIL WLKGSQRVPE EPGEQPIYMN FSEPLTKDMA T
CD300 molecule-like family member b (CD300LB), also known as CLM-7, is a gene located on chromosome 17q25.1, encoding an immunoreceptor that belongs to the CD300 family of molecules. CD300LB is primarily expressed on the surface of myeloid cells, including dendritic cells and macrophages, where it plays an essential role in modulating immune responses. The receptor is involved in the regulation of cell activation and inflammatory responses through its interactions with phosphatidylserine and other lipid ligands, which can lead to either activation or inhibition of immune cell functions. CD300LB influences processes like antigen presentation, phagocytosis, and cytokine production, thereby impacting innate immune responses and the maintenance of immune homeostasis. Its expression and activity are of particular interest in contexts like autoimmunity, infection, and cancer, where it may contribute to disease progression and serve as a potential therapeutic target in modulating immune functions.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD300 antigen like family member B; CD300 antigen-like family member B; CD300 molecule-like family member b; CD300b; CLM-7; CMRF-35-like molecule-7; CMRF35-A2; CMRF35-like molecule 7; immune activating receptor CD300b; Immune receptor expressed on myeloid cells 3; IREM-3; Leukocyte mono-Ig-like receptor 5; TREM-5; Triggering receptor expressed on myeloid cells 5
Gene Aliases: CD300B; CD300LB; CLM-7; CLM7; CMRF35-A2; CMRF35A2; Gm253; IREM-3; IREM3; LMIR5; TREM-5; TREM5; UNQ2530/PRO6029
UniProt ID: (Human) A8K4G0, (Mouse) Q3U497
Entrez Gene ID: (Human) 124599, (Mouse) 217304, (Rat) 501738
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support