Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
The CMRF35 antigen, which was identified by reactivity with a monoclonal antibody, is present on monocytes, neutrophils, and some T and B lymphocytes (Jackson et al., 1992 [PubMed 1349532]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD300 antigen-like family member C; CD300c; CD300c antigen; CLM-6; CMRF-35; CMRF35 antigen; CMRF35 leukocyte immunoglobulin-like receptor; CMRF35-A1; CMRF35-like molecule 6; CMRF35A leukocyte immunoglobulin-like receptor; Gcom1; IgSF16; Immunoglobulin superfamily member 16
Gene Aliases: CD300C; CLM-6; CMRF-35; CMRF-35A; CMRF35; CMRF35-A1; CMRF35A; CMRF35A1; IGSF16; LIR
UniProt ID: (Human) Q08708
Entrez Gene ID: (Human) 10871
Molecular Function: immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support