Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: MISIHNEEEN AFILDTLKKQ WKGPDDILLG MFYDTDDASF KWFDNSNMTF DKWTDQDDDE DLVDTCAFLH IKTGEWKKGN CEVSSVEGTL CKTAIPYKRK YLSDNHILIS ALVIASTVIL TVLGAIIWFL YKKHSDSRFT TVFSTAPQSP YNEDCVLVVG EENEYPVQFD
CD302 is a C-type lectin receptor involved in cell adhesion and migration, as well as endocytosis and phagocytosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type lectin BIMLEC; C-type lectin domain family 13 member A; C-type lectin domain family 13, member A; CD302; CD302 antigen; DEC205-associated C-type lectin 1; Type I transmembrane C-type lectin receptor DCL-1
Gene Aliases: BIMLEC; CD302; CLEC13A; DCL-1; DCL1; KIAA0022
UniProt ID: (Human) Q8IX05
Entrez Gene ID: (Human) 9936
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support