Product References
Transcriptomics and translatomics identify a robust inflammatory gene signature in brain endothelial cells after ischemic stroke.
Journal of neuroinflammation
Arbaizar-Rovirosa M,Gallizioli M,Lozano JJ,Sidorova J,Pedragosa J,Figuerola S,Chaparro-Cabanillas N,Boya P,Graupera M,Claret M,Urra X,Planas AM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Sep 11 00:00:00 EDT 2023
The Wnt/TCF7L1 transcriptional repressor axis drives primitive endoderm formation by antagonizing naive and formative pluripotency.
Nature communications
Athanasouli P,Balli M,De Jaime-Soguero A,Boel A,Papanikolaou S,van der Veer BK,Janiszewski A,Vanhessche T,Francis A,El Laithy Y,Nigro AL,Aulicino F,Koh KP,Pasque V,Cosma MP,Verfaillie C,Zwijsen A,Heindryckx B,Nikolaou C,Lluis F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Mar 03 00:00:00 EST 2023
Isolation and in vitro characterization of murine young-adult long bone skeletal progenitors.
Frontiers in endocrinology
Loopmans S,Stockmans I,Carmeliet G,Stegen S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Aug 19 00:00:00 EDT 2022
Synovial Fibroblast Sialylation Regulates Cell Migration and Activation of Inflammatory Pathways in Arthritogenesis.
Frontiers in immunology
Wang Y,Pan P,Khan A,Çil Ç,Pineda MA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Apr 08 00:00:00 EDT 2022
Macrophage IL-1β promotes arteriogenesis by autocrine STAT3- and NF-κB-mediated transcription of pro-angiogenic VEGF-A.
Cell reports
Mantsounga CS,Lee C,Neverson J,Sharma S,Healy A,Berus JM,Parry C,Ceneri NM,López-Giráldez F,Chun HJ,Lu Q,Sellke F,Choudhary G,Morrison AR
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Immunohistochemistry
Tue Feb 01 00:00:00 EST 2022
Lamin A/C-dependent chromatin architecture safeguards naïve pluripotency to prevent aberrant cardiovascular cell fate and function.
Nature communications
Wang Y,Elsherbiny A,Kessler L,Cordero J,Shi H,Serke H,Lityagina O,Trogisch FA,Mohammadi MM,El-Battrawy I,Backs J,Wieland T,Heineke J,Dobreva G
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Nov 04 00:00:00 EDT 2022
Fibroblast activation protein activated antifibrotic peptide delivery attenuates fibrosis in mouse models of liver fibrosis.
Nature communications
Lee J,Byun J,Shim G,Oh YK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Immunohistochemistry
Mon Mar 21 00:00:00 EDT 2022
Age-related changes in the local milieu of inflamed tissues cause aberrant neutrophil trafficking and subsequent remote organ damage.
Immunity
Barkaway A,Rolas L,Joulia R,Bodkin J,Lenn T,Owen-Woods C,Reglero-Real N,Stein M,Vázquez-Martínez L,Girbl T,Poston RN,Golding M,Saleeb RS,Thiriot A,von Andrian UH,Duchene J,Voisin MB,Bishop CL,Voehringer D,Roers A,Rot A,Lämmermann T,Nourshargh S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Immunocytochemistry
Tue Jul 13 00:00:00 EDT 2021
Fli1+ cells transcriptional analysis reveals an Lmo2-Prdm16 axis in angiogenesis.
Proceedings of the National Academy of Sciences of the United States of America
Matrone G,Xia B,Chen K,Denvir MA,Baker AH,Cooke JP
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Immunocytochemistry
Tue Aug 03 00:00:00 EDT 2021
Photoreceptor protection by mesenchymal stem cell transplantation identifies exosomal MiR-21 as a therapeutic for retinal degeneration.
Cell death and differentiation
Deng CL,Hu CB,Ling ST,Zhao N,Bao LH,Zhou F,Xiong YC,Chen T,Sui BD,Yu XR,Hu CH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Mar 01 00:00:00 EST 2021
Anti-adipogenic signals at the onset of obesity-related inflammation in white adipose tissue.
Cellular and molecular life sciences : CMLS
Caputo T,Tran VDT,Bararpour N,Winkler C,Aguileta G,Trang KB,Giordano Attianese GMP,Wilson A,Thomas A,Pagni M,Guex N,Desvergne B,Gilardi F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
SORLA is required for insulin-induced expansion of the adipocyte precursor pool in visceral fat.
The Journal of cell biology
Schmidt V,Horváth C,Dong H,Blüher M,Qvist P,Wolfrum C,Willnow TE
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Dec 06 00:00:00 EST 2021
Galectin-3 Enhances Vascular Endothelial Growth Factor-A Receptor 2 Activity in the Presence of Vascular Endothelial Growth Factor.
Frontiers in cell and developmental biology
Cano I,Hu Z,AbuSamra DB,Saint-Geniez M,Ng YSE,Argüeso P,D'Amore PA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Western Blot
Sat Jul 16 00:00:00 EDT 2022
Tendon and motor phenotypes in the mouse model of recessive osteogenesis imperfecta.
eLife
Grol MW,Haelterman NA,Lim J,Munivez EM,Archer M,Hudson DM,Tufa SF,Keene DR,Lei K,Park D,Kuzawa CD,Ambrose CG,Eyre DR,Lee BH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Wed May 26 00:00:00 EDT 2021
The antitumoral effects of chemerin are independent from leukocyte recruitment and mediated by inhibition of neoangiogenesis.
Oncotarget
Dubois-Vedrenne I,Al Delbany D,De Henau O,Robert V,Vernimmen M,Langa F,Lefort A,Libert F,Wittamer V,Parmentier M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Immunocytochemistry
Tue Sep 14 00:00:00 EDT 2021
GRHL3 activates FSCN1 to relax cell-cell adhesions between migrating keratinocytes during wound reepithelialization.
JCI insight
Kashgari G,Venkatesh S,Refuerzo S,Pham B,Bayat A,Klein RH,Ramos R,Ta AP,Plikus MV,Wang PH,Andersen B
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Wed Sep 08 00:00:00 EDT 2021
The IRE1/XBP1 signaling axis promotes skeletal muscle regeneration through a cell non-autonomous mechanism.
eLife
Roy A,Tomaz da Silva M,Bhat R,Bohnert KR,Iwawaki T,Kumar A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Nov 23 00:00:00 EST 2021
Interleukin-11-expressing fibroblasts have a unique gene signature correlated with poor prognosis of colorectal cancer.
Nature communications
Nishina T,Deguchi Y,Ohshima D,Takeda W,Ohtsuka M,Shichino S,Ueha S,Yamazaki S,Kawauchi M,Nakamura E,Nishiyama C,Kojima Y,Adachi-Akahane S,Hasegawa M,Nakayama M,Oshima M,Yagita H,Shibuya K,Mikami T,Inohara N,Matsushima K,Tada N,Nakano H
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Apr 16 00:00:00 EDT 2021
Therapeutic Potential of Mesenchymal Stem Cells in a Pre-Clinical Model of Diabetic Kidney Disease and Obesity.
International journal of molecular sciences
Sávio-Silva C,Soinski-Sousa PE,Simplício-Filho A,Bastos RMC,Beyerstedt S,Rangel ÉB
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Thu Feb 04 00:00:00 EST 2021
Protocols for isolation and characterization of mouse placental hemogenic endothelial cells.
STAR protocols
Liang G,Huang B,Wang F,Liu F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Dec 17 00:00:00 EST 2021
Knockout RAGE alleviates cardiac fibrosis through repressing endothelial-to-mesenchymal transition (EndMT) mediated by autophagy.
Cell death & disease
Zhang L,He J,Wang J,Liu J,Chen Z,Deng B,Wei L,Wu H,Liang B,Li H,Huang Y,Lu L,Yang Z,Xian S,Wang L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue May 11 00:00:00 EDT 2021
Loss of α2-6 sialylation promotes the transformation of synovial fibroblasts into a pro-inflammatory phenotype in arthritis.
Nature communications
Wang Y,Khan A,Antonopoulos A,Bouché L,Buckley CD,Filer A,Raza K,Li KP,Tolusso B,Gremese E,Kurowska-Stolarska M,Alivernini S,Dell A,Haslam SM,Pineda MA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Apr 20 00:00:00 EDT 2021
Mice with induced pulmonary morbidities display severe lung inflammation and mortality following exposure to SARS-CoV-2.
JCI insight
Falach R,Bar-On L,Lazar S,Kadar T,Mazor O,Aftalion M,Gur D,Evgy Y,Shifman O,Aminov T,Israeli O,Cohen-Gihon I,Zaide G,Gutman H,Vagima Y,Makdasi E,Stein D,Rosenfeld R,Alcalay R,Zahavy E,Levy H,Glinert I,Ben-Shmuel A,Israely T,Melamed S,Politi B,Achdout H,Yitzhaki S,Kronman C,Sabo T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Jun 22 00:00:00 EDT 2021
SPARC regulation of PMN clearance protects from pristane-induced lupus and rheumatoid arthritis.
iScience
Sangaletti S,Botti L,Gulino A,Lecis D,Bassani B,Portararo P,Milani M,Cancila V,De Cecco L,Dugo M,Tripodo C,Colombo MP
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Jun 25 00:00:00 EDT 2021
Deficiency of Dietary Fiber Modulates Gut Microbiota Composition, Neutrophil Recruitment and Worsens Experimental Colitis.
Frontiers in immunology
Shen S,Prame Kumar K,Wen SW,Shim R,Wanrooy BJ,Stanley D,Moore RJ,Van TTH,Robert R,Hickey MJ,Wong CHY
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Immunohistochemistry
Mon Jul 05 00:00:00 EDT 2021
Protocols for endothelial cell isolation from mouse tissues: brain, choroid, lung, and muscle.
STAR protocols
Conchinha NV,Sokol L,Teuwen LA,Veys K,Dumas SJ,Meta E,García-Caballero M,Geldhof V,Chen R,Treps L,Borri M,de Zeeuw P,Falkenberg KD,Dubois C,Parys M,de Rooij LPMH,Rohlenova K,Goveia J,Schoonjans L,Dewerchin M,Eelen G,Li X,Kalucka J,Carmeliet P
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Sep 17 00:00:00 EDT 2021
Testosterone metabolites differentially regulate obesogenesis and fat distribution.
Molecular metabolism
Sebo ZL,Rodeheffer MS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Meningeal lymphatic vessels regulate brain tumor drainage and immunity.
Cell research
Hu X,Deng Q,Ma L,Li Q,Chen Y,Liao Y,Zhou F,Zhang C,Shao L,Feng J,He T,Ning W,Kong Y,Huo Y,He A,Liu B,Zhang J,Adams R,He Y,Tang F,Bian X,Luo J
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
Therapy-Induced Senescence Drives Bone Loss.
Cancer research
Yao Z,Murali B,Ren Q,Luo X,Faget DV,Cole T,Ricci B,Thotala D,Monahan J,van Deursen JM,Baker D,Faccio R,Schwarz JK,Stewart SA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
Membrane expression of the estrogen receptor ERα is required for intercellular communications in the mammary epithelium.
Development (Cambridge, England)
Gagniac L,Rusidzé M,Boudou F,Cagnet S,Adlanmerini M,Jeannot P,Gaide N,Giton F,Besson A,Weyl A,Gourdy P,Raymond-Letron I,Arnal JF,Brisken C,Lenfant F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Wed Mar 11 00:00:00 EDT 2020
HSPA12B Secreted by Tumor-Associated Endothelial Cells Might Induce M2 Polarization of Macrophages via Activating PI3K/Akt/mTOR Signaling.
OncoTargets and therapy
Zhou J,Zhang A,Fan L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Immunocytochemistry
Sun Apr 17 00:00:00 EDT 2022
Defining the role of pulmonary endothelial cell heterogeneity in the response to acute lung injury.
eLife
Niethamer TK,Stabler CT,Leach JP,Zepp JA,Morley MP,Babu A,Zhou S,Morrisey EE
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Feb 24 00:00:00 EST 2020
Enhancer remodeling promotes tumor-initiating activity in NRF2-activated non-small cell lung cancers.
Nature communications
Okazaki K,Anzawa H,Liu Z,Ota N,Kitamura H,Onodera Y,Alam MM,Matsumaru D,Suzuki T,Katsuoka F,Tadaka S,Motoike I,Watanabe M,Hayasaka K,Sakurada A,Okada Y,Yamamoto M,Suzuki T,Kinoshita K,Sekine H,Motohashi H
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Nov 20 00:00:00 EST 2020
Snail promotes the generation of vascular endothelium by breast cancer cells.
Cell death & disease
Chang Z,Zhang Y,Liu J,Zheng Y,Li H,Kong Y,Li P,Peng H,Shi Y,Cao B,Ran F,Chen Y,Song Y,Ye Q,Ding L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Jun 15 00:00:00 EDT 2020
Sierra: discovery of differential transcript usage from polyA-captured single-cell RNA-seq data.
Genome biology
Patrick R,Humphreys DT,Janbandhu V,Oshlack A,Ho JWK,Harvey RP,Lo KK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Wed Jul 08 00:00:00 EDT 2020
Cardiac fibroblast proliferation rates and collagen expression mature early and are unaltered with advancing age.
JCI insight
Wu R,Ma F,Tosevska A,Farrell C,Pellegrini M,Deb A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Thu Dec 17 00:00:00 EST 2020
Myh11+ microvascular mural cells and derived mesenchymal stem cells promote retinal fibrosis.
Scientific reports
Ray HC,Corliss BA,Bruce AC,Kesting S,Dey P,Mansour J,Seaman SA,Smolko CM,Mathews C,Dey BK,Owens GK,Peirce SM,Yates PA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Sep 25 00:00:00 EDT 2020
Diaphanous-related formin mDia2 regulates beta2 integrins to control hematopoietic stem and progenitor cell engraftment.
Nature communications
Mei Y,Han X,Liu Y,Yang J,Sumagin R,Ji P
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Immunohistochemistry
Tue Jun 23 00:00:00 EDT 2020
Exosomes derived from GDNF-modified human adipose mesenchymal stem cells ameliorate peritubular capillary loss in tubulointerstitial fibrosis by activating the SIRT1/eNOS signaling pathway.
Theranostics
Chen L,Wang Y,Li S,Zuo B,Zhang X,Wang F,Sun D
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Immunocytochemistry
Mon May 24 00:00:00 EDT 2021
Apelin inhibition prevents resistance and metastasis associated with anti-angiogenic therapy.
EMBO molecular medicine
Uribesalgo I,Hoffmann D,Zhang Y,Kavirayani A,Lazovic J,Berta J,Novatchkova M,Pai TP,Wimmer RA,László V,Schramek D,Karim R,Tortola L,Deswal S,Haas L,Zuber J,Szűcs M,Kuba K,Dome B,Cao Y,Haubner BJ,Penninger JM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Thu Aug 01 00:00:00 EDT 2019
Vascular endothelial growth factor pathway promotes osseointegration and CD31hiEMCNhi endothelium expansion in a mouse tibial implant model: an animal study.
The bone & joint journal
Ji G,Xu R,Niu Y,Li N,Ivashkiv L,Bostrom MPG,Greenblatt MB,Yang X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2019
LPS‑induced upregulation of the TLR4 signaling pathway inhibits osteogenic differentiation of human periodontal ligament stem cells under inflammatory conditions.
International journal of molecular medicine
Yu B,Li Q,Zhou M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Sat Jun 01 00:00:00 EDT 2019
Cell-specific ablation of Hsp47 defines the collagen-producing cells in the injured heart.
JCI insight
Khalil H,Kanisicak O,Vagnozzi RJ,Johansen AK,Maliken BD,Prasad V,Boyer JG,Brody MJ,Schips T,Kilian KK,Correll RN,Kawasaki K,Nagata K,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Thu Aug 08 00:00:00 EDT 2019
Single-cell expression profiling reveals dynamic flux of cardiac stromal, vascular and immune cells in health and injury.
eLife
Farbehi N,Patrick R,Dorison A,Xaymardan M,Janbandhu V,Wystub-Lis K,Ho JW,Nordon RE,Harvey RP
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Mar 26 00:00:00 EDT 2019
PDGFRα+ stromal adipocyte progenitors transition into epithelial cells during lobulo-alveologenesis in the murine mammary gland.
Nature communications
Joshi PA,Waterhouse PD,Kasaian K,Fang H,Gulyaeva O,Sul HS,Boutros PC,Khokha R
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Apr 15 00:00:00 EDT 2019
Growth Factor Screening in Dystrophic Muscles Reveals PDGFB/PDGFRB-Mediated Migration of Interstitial Stem Cells.
International journal of molecular sciences
Camps J,Grosemans H,Gijsbers R,Maes C,Sampaolesi M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Mar 05 00:00:00 EST 2019
Metastatic cancers promote cachexia through ZIP14 upregulation in skeletal muscle.
Nature medicine
Wang G,Biswas AK,Ma W,Kandpal M,Coker C,Grandgenett PM,Hollingsworth MA,Jain R,Tanji K,Lόpez-Pintado S,Borczuk A,Hebert D,Jenkitkasemwong S,Hojyo S,Davuluri RV,Knutson MD,Fukada T,Acharyya S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Targeting skeletal endothelium to ameliorate bone loss.
Nature medicine
Xu R,Yallowitz A,Qin A,Wu Z,Shin DY,Kim JM,Debnath S,Ji G,Bostrom MP,Yang X,Zhang C,Dong H,Kermani P,Lalani S,Li N,Liu Y,Poulos MG,Wach A,Zhang Y,Inoue K,Di Lorenzo A,Zhao B,Butler JM,Shim JH,Glimcher LH,Greenblatt MB
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Endothelial cells in the innate response to allergens and initiation of atopic asthma.
The Journal of clinical investigation
Asosingh K,Weiss K,Queisser K,Wanner N,Yin M,Aronica M,Erzurum S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Jul 02 00:00:00 EDT 2018
Indian Hedgehog Suppresses a Stromal Cell-Driven Intestinal Immune Response.
Cellular and molecular gastroenterology and hepatology
Westendorp BF,Büller NVJA,Karpus ON,van Dop WA,Koster J,Versteeg R,Koelink PJ,Snel CY,Meisner S,Roelofs JJTH,Uhmann A,Ver Loren van Themaat E,Heijmans J,Hahn H,Muncan V,Wildenberg ME,van den Brink GR
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Thu Mar 31 00:00:00 EDT 2022
Islr regulates canonical Wnt signaling-mediated skeletal muscle regeneration by stabilizing Dishevelled-2 and preventing autophagy.
Nature communications
Zhang K,Zhang Y,Gu L,Lan M,Liu C,Wang M,Su Y,Ge M,Wang T,Yu Y,Liu C,Li L,Li Q,Zhao Y,Yu Z,Wang F,Li N,Meng Q
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Dec 03 00:00:00 EST 2018
Adipocyte hypertrophy and lipid dynamics underlie mammary gland remodeling after lactation.
Nature communications
Zwick RK,Rudolph MC,Shook BA,Holtrup B,Roth E,Lei V,Van Keymeulen A,Seewaldt V,Kwei S,Wysolmerski J,Rodeheffer MS,Horsley V
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Sep 04 00:00:00 EDT 2018
ISRE-Reporter Mouse Reveals High Basal and Induced Type I IFN Responses in Inflammatory Monocytes.
Cell reports
Uccellini MB,García-Sastre A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Dec 04 00:00:00 EST 2018
Stem cell factor is selectively secreted by arterial endothelial cells in bone marrow.
Nature communications
Xu C,Gao X,Wei Q,Nakahara F,Zimmerman SE,Mar J,Frenette PS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Jun 22 00:00:00 EDT 2018
Age-specific biological and molecular profiling distinguishes paediatric from adult acute myeloid leukaemias.
Nature communications
Chaudhury S,O'Connor C,Cañete A,Bittencourt-Silvestre J,Sarrou E,Prendergast Á,Choi J,Johnston P,Wells CA,Gibson B,Keeshan K
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Dec 11 00:00:00 EST 2018
Gpr124 is essential for blood-brain barrier integrity in central nervous system disease.
Nature medicine
Chang J,Mancuso MR,Maier C,Liang X,Yuki K,Yang L,Kwong JW,Wang J,Rao V,Vallon M,Kosinski C,Zhang JJ,Mah AT,Xu L,Li L,Gholamin S,Reyes TF,Li R,Kuhnert F,Han X,Yuan J,Chiou SH,Brettman AD,Daly L,Corney DC,Cheshier SH,Shortliffe LD,Wu X,Snyder M,Chan P,Giffard RG,Chang HY,Andreasson K,Kuo CJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
Tumour ischaemia by interferon-γ resembles physiological blood vessel regression.
Nature
Kammertoens T,Friese C,Arina A,Idel C,Briesemeister D,Rothe M,Ivanov A,Szymborska A,Patone G,Kunz S,Sommermeyer D,Engels B,Leisegang M,Textor A,Fehling HJ,Fruttiger M,Lohoff M,Herrmann A,Yu H,Weichselbaum R,Uckert W,Hübner N,Gerhardt H,Beule D,Schreiber H,Blankenstein T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Thu May 04 00:00:00 EDT 2017
Increased Vascular Permeability in the Bone Marrow Microenvironment Contributes to Disease Progression and Drug Response in Acute Myeloid Leukemia.
Cancer cell
Passaro D,Di Tullio A,Abarrategi A,Rouault-Pierre K,Foster K,Ariza-McNaughton L,Montaner B,Chakravarty P,Bhaw L,Diana G,Lassailly F,Gribben J,Bonnet D
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Sep 11 00:00:00 EDT 2017
Preexisting endothelial cells mediate cardiac neovascularization after injury.
The Journal of clinical investigation
He L,Huang X,Kanisicak O,Li Y,Wang Y,Li Y,Pu W,Liu Q,Zhang H,Tian X,Zhao H,Liu X,Zhang S,Nie Y,Hu S,Miao X,Wang QD,Wang F,Chen T,Xu Q,Lui KO,Molkentin JD,Zhou B
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
Obesity reversibly depletes the basal cell population and enhances mammary epithelial cell estrogen receptor alpha expression and progenitor activity.
Breast cancer research : BCR
Chamberlin T,D'Amato JV,Arendt LM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Wed Nov 29 00:00:00 EST 2017
Mesenchymal Stem Cell Therapy Protects Lungs from Radiation-Induced Endothelial Cell Loss by Restoring Superoxide Dismutase 1 Expression.
Antioxidants & redox signaling
Klein D,Steens J,Wiesemann A,Schulz F,Kaschani F,Röck K,Yamaguchi M,Wirsdörfer F,Kaiser M,Fischer JW,Stuschke M,Jendrossek V
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Apr 10 00:00:00 EDT 2017
Estrogen modulates mesenchyme-epidermis interactions in the adult nipple.
Development (Cambridge, England)
Wu HJ,Oh JW,Spandau DF,Tholpady S,Diaz J,Schroeder LJ,Offutt CD,Glick AB,Plikus MV,Koyama S,Foley J
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Sat Apr 15 00:00:00 EDT 2017
Characterization of transgene expression and pDNA distribution of the suctioned kidney in mice.
Drug delivery
Oyama N,Fuchigami Y,Fumoto S,Sato M,Hagimori M,Shimizu K,Kawakami S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Immunocytochemistry
Wed Nov 01 00:00:00 EDT 2017
Cornea lymphatics drive the CD8+ T-cell response to herpes simplex virus-1.
Immunology and cell biology
Gurung HR,Carr MM,Carr DJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Sun Jan 01 00:00:00 EST 2017
β3-Adrenergic Regulation of EPC Features Through Manipulation of the Bone Marrow MSC Niche.
Journal of cellular biochemistry
Vafaei R,Nassiri SM,Siavashi V
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2017
Dynamic Patterns of Clonal Evolution in Tumor Vasculature Underlie Alterations in Lymphocyte-Endothelial Recognition to Foster Tumor Immune Escape.
Cancer research
Corey DM,Rinkevich Y,Weissman IL
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Mar 15 00:00:00 EDT 2016
Endothelial cell survival during angiogenesis requires the pro-survival protein MCL1.
Cell death and differentiation
Watson EC,Whitehead L,Adams RH,Dewson G,Coultas L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Lack of VEGFR2 signaling causes maldevelopment of the intestinal microvasculature and facilitates necrotizing enterocolitis in neonatal mice.
American journal of physiology. Gastrointestinal and liver physiology
Yan X,Managlia E,Liu SX,Tan XD,Wang X,Marek C,De Plaen IG
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Sun May 01 00:00:00 EDT 2016
A cell-autonomous tumour suppressor role of RAF1 in hepatocarcinogenesis.
Nature communications
Jeric I,Maurer G,Cavallo AL,Raguz J,Desideri E,Tarkowski B,Parrini M,Fischer I,Zatloukal K,Baccarini M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Wed Dec 21 00:00:00 EST 2016
Genetic lineage tracing defines myofibroblast origin and function in the injured heart.
Nature communications
Kanisicak O,Khalil H,Ivey MJ,Karch J,Maliken BD,Correll RN,Brody MJ,J Lin SC,Aronow BJ,Tallquist MD,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Jul 22 00:00:00 EDT 2016
BMP and Hedgehog Regulate Distinct AGM Hematopoietic Stem Cells Ex Vivo.
Stem cell reports
Crisan M,Solaimani Kartalaei P,Neagu A,Karkanpouna S,Yamada-Inagawa T,Purini C,Vink CS,van der Linden R,van Ijcken W,Chuva de Sousa Lopes SM,Monteiro R,Mummery C,Dzierzak E
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Mar 08 00:00:00 EST 2016
Self-renewing diploid Axin2(+) cells fuel homeostatic renewal of the liver.
Nature
Wang B,Zhao L,Fish M,Logan CY,Nusse R
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Thu Aug 13 00:00:00 EDT 2015
Host microbiota constantly control maturation and function of microglia in the CNS.
Nature neuroscience
Erny D,Hrabě de Angelis AL,Jaitin D,Wieghofer P,Staszewski O,David E,Keren-Shaul H,Mahlakoiv T,Jakobshagen K,Buch T,Schwierzeck V,Utermöhlen O,Chun E,Garrett WS,McCoy KD,Diefenbach A,Staeheli P,Stecher B,Amit I,Prinz M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2015
Rapid depot-specific activation of adipocyte precursor cells at the onset of obesity.
Nature cell biology
Jeffery E,Church CD,Holtrup B,Colman L,Rodeheffer MS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
Vascular endothelial growth factor c/vascular endothelial growth factor receptor 3 signaling regulates chemokine gradients and lymphocyte migration from tissues to lymphatics.
Transplantation
Iwami D,Brinkman CC,Bromberg JS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
Conditional immortalization of primary adipocyte precursor cells.
Adipocyte
Church C,Brown M,Rodeheffer MS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Aug 10 00:00:00 EDT 2015
Targeted Gene Correction in Osteopetrotic-Induced Pluripotent Stem Cells for the Generation of Functional Osteoclasts.
Stem cell reports
Neri T,Muggeo S,Paulis M,Caldana ME,Crisafulli L,Strina D,Focarelli ML,Faggioli F,Recordati C,Scaramuzza S,Scanziani E,Mantero S,Buracchi C,Sobacchi C,Lombardo A,Naldini L,Vezzoni P,Villa A,Ficara F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Oct 13 00:00:00 EDT 2015
Rejuvenation of the muscle stem cell population restores strength to injured aged muscles.
Nature medicine
Cosgrove BD,Gilbert PM,Porpiglia E,Mourkioti F,Lee SP,Corbel SY,Llewellyn ME,Delp SL,Blau HM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Sat Mar 01 00:00:00 EST 2014
c-kit+ cells minimally contribute cardiomyocytes to the heart.
Nature
van Berlo JH,Kanisicak O,Maillet M,Vagnozzi RJ,Karch J,Lin SC,Middleton RC,Marbán E,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Thu May 15 00:00:00 EDT 2014
Mesenchymal-endothelial transition contributes to cardiac neovascularization.
Nature
Ubil E,Duan J,Pillai IC,Rosa-Garrido M,Wu Y,Bargiacchi F,Lu Y,Stanbouly S,Huang J,Rojas M,Vondriska TM,Stefani E,Deb A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Thu Oct 30 00:00:00 EDT 2014
Cell-autonomous heparanase modulates self-renewal and migration in bone marrow-derived mesenchymal stem cells.
Journal of biomedical science
Cheng CC,Lee YH,Lin SP,Huangfu WC,Liu IH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Thu Mar 13 00:00:00 EDT 2014
Characterization of Cre recombinase activity for in vivo targeting of adipocyte precursor cells.
Stem cell reports
Krueger KC,Costa MJ,Du H,Feldman BJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Tue Dec 09 00:00:00 EST 2014
miR-126 and miR-126* repress recruitment of mesenchymal stem cells and inflammatory monocytes to inhibit breast cancer metastasis.
Nature cell biology
Zhang Y,Yang P,Sun T,Li D,Xu X,Rui Y,Li C,Chong M,Ibrahim T,Mercatali L,Amadori D,Lu X,Xie D,Li QJ,Wang XF
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
The expression of Sox17 identifies and regulates haemogenic endothelium.
Nature cell biology
Clarke RL,Yzaguirre AD,Yashiro-Ohtani Y,Bondue A,Blanpain C,Pear WS,Speck NA,Keller G
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Wed May 01 00:00:00 EDT 2013
Etv2 rescues Flk1 mutant embryoid bodies.
Genesis (New York, N.Y. : 2000)
Rasmussen TL,Martin CM,Walter CA,Shi X,Perlingeiro R,Koyano-Nakagawa N,Garry DJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2013
Mouse models and techniques for the isolation of the diabetic endothelium.
ISRN endocrinology
Darrow AL,Maresh JG,Shohet RV
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Wed Jul 10 00:00:00 EDT 2013
Endothelial cells provide an instructive niche for the differentiation and functional polarization of M2-like macrophages.
Blood
He H,Xu J,Warren CM,Duan D,Li X,Wu L,Iruela-Arispe ML
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Thu Oct 11 00:00:00 EDT 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2012
Intramuscular VEGF activates an SDF1-dependent progenitor cell cascade and an SDF1-independent muscle paracrine cascade for cardiac repair.
American journal of physiology. Heart and circulatory physiology
Zisa D,Shabbir A,Mastri M,Taylor T,Aleksic I,McDaniel M,Suzuki G,Lee T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2011
Heart failure therapy mediated by the trophic activities of bone marrow mesenchymal stem cells: a noninvasive therapeutic regimen.
American journal of physiology. Heart and circulatory physiology
Shabbir A,Zisa D,Suzuki G,Lee T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2009
Intramuscular VEGF repairs the failing heart: role of host-derived growth factors and mobilization of progenitor cells.
American journal of physiology. Regulatory, integrative and comparative physiology
Zisa D,Shabbir A,Mastri M,Suzuki G,Lee T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 15-0311-82) in Flow Cytometry
Sun Nov 01 00:00:00 EDT 2009