Product References
A novel approach for transforming breast cancer stem cells into endothelial cells.
Experimental and therapeutic medicine
Mao QQ,Ji XC,Zhang JN,Teng WF,Zhou SC
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Thu Feb 01 00:00:00 UTC 2024
Tumor-derived mesenchymal progenitor cell-related genes in the regulation of breast cancer proliferation.
Gland surgery
Chen Y,Zhu L,Wang Y,Hu J,Zhang H,Zhu J,Gong W,Liu X,Xiao F,Li X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Wed Mar 27 00:00:00 UTC 2024
Epigenetic landscape reveals MECOM as an endothelial lineage regulator.
Nature communications
Lv J,Meng S,Gu Q,Zheng R,Gao X,Kim JD,Chen M,Xia B,Zuo Y,Zhu S,Zhao D,Li Y,Wang G,Wang X,Meng Q,Cao Q,Cooke JP,Fang L,Chen K,Zhang L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Tue Apr 25 00:00:00 UTC 2023
MiR-451a and let-7i-5p loaded extracellular vesicles attenuate heme-induced inflammation in hiPSC-derived endothelial cells.
Frontiers in immunology
Thomas JJ,Harp KO,Bashi A,Hood JL,Botchway F,Wilson MD,Thompson WE,Stiles JK,Driss A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Wed Jan 11 00:00:00 UTC 2023
Angiogenic stem cell delivery platform to augment post-infarction neovasculature and reverse ventricular remodeling.
Scientific reports
Shin HS,Thakore A,Tada Y,Pedroza AJ,Ikeda G,Chen IY,Chan D,Jaatinen KJ,Yajima S,Pfrender EM,Kawamura M,Yang PC,Wu JC,Appel EA,Fischbein MP,Woo Y,Shudo Y
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Thu Oct 20 00:00:00 UTC 2022
The Composition of Adipose-Derived Regenerative Cells Isolated from Lipoaspirate Using a Point of Care System Does Not Depend on the Subject's Individual Age, Sex, Body Mass Index and Ethnicity.
Cells
Schmitz C,Alt C,Azares AR,Pearce DA,Facile TR,Furia JP,Maffulli N,Huang C,Alt EU
12-0319-42 was used in Flow cytometry/Cell sorting to show that key characteristics of unmodified, autologous, adipose-derived regenerative cells (UA-ADRCs) can be independent of the subject's age, sex, BMI and ethnicity.
Wed Dec 21 00:00:00 UTC 2022
Expression and role of interleukin-1β and associated biomarkers in deep vein thrombosis.
Experimental and therapeutic medicine
Pai RZ,Fang Q,Tian G,Zhu B,Ge X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Wed Dec 01 00:00:00 UTC 2021
Injectable hydrogel with MSNs/microRNA-21-5p delivery enables both immunomodification and enhanced angiogenesis for myocardial infarction therapy in pigs.
Science advances
Li Y,Chen X,Jin R,Chen L,Dang M,Cao H,Dong Y,Cai B,Bai G,Gooding JJ,Liu S,Zou D,Zhang Z,Yang C
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2021
A Human Skeletal Muscle Atlas Identifies the Trajectories of Stem and Progenitor Cells across Development and from Human Pluripotent Stem Cells.
Cell stem cell
Xi H,Langerman J,Sabri S,Chien P,Young CS,Younesi S,Hicks M,Gonzalez K,Fujiwara W,Marzi J,Liebscher S,Spencer M,Van Handel B,Evseenko D,Schenke-Layland K,Plath K,Pyle AD
12-0319-42 was used in Flow Cytometry to profile human embryonic, fetal and postnatal skeletal muscle tissues to develop a roadmap of muscle ontogeny.
Thu Jul 02 00:00:00 UTC 2020
Placental endovascular extravillous trophoblasts (enEVTs) educate maternal T-cell differentiation along the maternal-placental circulation.
Cell proliferation
Ma Y,Yang Q,Fan M,Zhang L,Gu Y,Jia W,Li Z,Wang F,Li YX,Wang J,Li R,Shao X,Wang YL
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Fri May 01 00:00:00 UTC 2020
Placental endovascular extravillous trophoblasts (enEVTs) educate maternal T-cell differentiation along the maternal-placental circulation.
Cell proliferation
Ma Y,Yang Q,Fan M,Zhang L,Gu Y,Jia W,Li Z,Wang F,Li YX,Wang J,Li R,Shao X,Wang YL
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Fri May 01 00:00:00 UTC 2020
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Mon Sep 28 00:00:00 UTC 2020
Aging affects responsiveness of peripheral blood mononuclear cells to immunosuppression of periodontal ligament stem cells.
The Journal of international medical research
Li X,Zhang B,Wang H,Zhao X,Zhang Z,Ding G,Wei F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Fri Apr 03 00:00:00 UTC 2020
Developing Human Skin Contains Lymphocytes Demonstrating a Memory Signature.
Cell reports. Medicine
Dhariwala MO,Karthikeyan D,Vasquez KS,Farhat S,Weckel A,Taravati K,Leitner EG,Clancy S,Pauli M,Piper ML,Cohen JN,Ashouri JF,Lowe MM,Rosenblum MD,Scharschmidt TC
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Tue Nov 17 00:00:00 UTC 2020
Both Human Hematoma Punctured from Pelvic Fractures and Serum Increase Muscle Resident Stem Cells Response to BMP9: A Multivariate Statistical Approach.
Journal of clinical medicine
Alinejad Y,Lauzon MA,Grenier G,Balg F,Faucheux N
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Sun Apr 19 00:00:00 UTC 2020
Functionally specialized human CD4+ T-cell subsets express physicochemically distinct TCRs.
eLife
Kasatskaya SA,Ladell K,Egorov ES,Miners KL,Davydov AN,Metsger M,Staroverov DB,Matveyshina EK,Shagina IA,Mamedov IZ,Izraelson M,Shelyakin PV,Britanova OV,Price DA,Chudakov DM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Tue Dec 08 00:00:00 UTC 2020
Vascular Tumor Recapitulated in Endothelial Cells from hiPSCs Engineered to Express the SERPINE1-FOSB Translocation.
Cell reports. Medicine
van IJzendoorn DGP,Salvatori DCF,Cao X,van den Hil F,Briaire-de Bruijn IH,de Jong D,Mei H,Mummery CL,Szuhai K,Bovée JVMG,Orlova VV
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Tue Dec 22 00:00:00 UTC 2020
Spatial and Single-Cell Transcriptional Profiling Identifies Functionally Distinct Human Dermal Fibroblast Subpopulations.
The Journal of investigative dermatology
Philippeos C,Telerman SB,Oulès B,Pisco AO,Shaw TJ,Elgueta R,Lombardi G,Driskell RR,Soldin M,Lynch MD,Watt FM
Sun Apr 01 00:00:00 UTC 2018
Expansion of functional personalized cells with specific transgene combinations.
Nature communications
Lipps C,Klein F,Wahlicht T,Seiffert V,Butueva M,Zauers J,Truschel T,Luckner M,Köster M,MacLeod R,Pezoldt J,Hühn J,Yuan Q,Müller PP,Kempf H,Zweigerdt R,Dittrich-Breiholz O,Pufe T,Beckmann R,Drescher W,Riancho J,Sañudo C,Korff T,Opalka B,Rebmann V,Göthert JR,Alves PM,Ott M,Schucht R,Hauser H,Wirth D,May T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Thu Mar 08 00:00:00 UTC 2018
Prostaglandin E2 Is Required for BMP4-Induced Mesoderm Differentiation of Human Embryonic Stem Cells.
Stem cell reports
Zhang B,He L,Liu Y,Zhang J,Zeng Q,Wang S,Fan Z,Fang F,Chen L,Lv Y,Xi J,Yue W,Li Y,Pei X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Tue Mar 13 00:00:00 UTC 2018
Genetic Architecture of Adaptive Immune System Identifies Key Immune Regulators.
Cell reports
Lagou V,Garcia-Perez JE,Smets I,Van Horebeek L,Vandebergh M,Chen L,Mallants K,Prezzemolo T,Hilven K,Humblet-Baron S,Moisse M,Van Damme P,Boeckxstaens G,Bowness P,Dubois B,Dooley J,Liston A,Goris A
12-0319-42 was used in Flow Cytometry to identify eight genome-wide significant associations explaining 6%-20% of variance in immune system regulation.
Tue Oct 16 00:00:00 UTC 2018
MMP-2 and MMP-14 Silencing Inhibits VEGFR2 Cleavage and Induces the Differentiation of Porcine Adipose-Derived Mesenchymal Stem Cells to Endothelial Cells.
Stem cells translational medicine
Almalki SG,Llamas Valle Y,Agrawal DK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Mon May 01 00:00:00 UTC 2017
ERK signaling is required for VEGF-A/VEGFR2-induced differentiation of porcine adipose-derived mesenchymal stem cells into endothelial cells.
Stem cell research & therapy
Almalki SG,Agrawal DK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Fri May 12 00:00:00 UTC 2017
TGF-β reduces DNA ds-break repair mechanisms to heighten genetic diversity and adaptability of CD44+/CD24- cancer cells.
eLife
Pal D,Pertot A,Shirole NH,Yao Z,Anaparthy N,Garvin T,Cox H,Chang K,Rollins F,Kendall J,Edwards L,Singh VA,Stone GC,Schatz MC,Hicks J,Hannon GJ,Sordella R
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Mon Jan 16 00:00:00 UTC 2017
Increased infiltration and tolerised antigen-specific CD8+ TEM cells in tumor but not peripheral blood have no impact on survival of HCMV+ glioblastoma patients.
Oncoimmunology
Bahador M,Gras Navarro A,Rahman MA,Dominguez-Valentin M,Sarowar S,Ulvestad E,Njølstad G,Lie SA,Kristoffersen EK,Bratland E,Chekenya M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Tue Jan 12 00:00:00 UTC 2021
Targeted Disruption of TCF12 Reveals HEB as Essential in Human Mesodermal Specification and Hematopoiesis.
Stem cell reports
Li Y,Brauer PM,Singh J,Xhiku S,Yoganathan K,Zúñiga-Pflücker JC,Anderson MK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Tue Sep 12 00:00:00 UTC 2017
Endothelial cells' biophysical, biochemical, and chromosomal aberrancies in high-glucose condition within the diabetic range.
Cell biochemistry and function
Rezabakhsh A,Nabat E,Yousefi M,Montazersaheb S,Cheraghi O,Mehdizadeh A,Fathi F,Movassaghpour AA,Maleki-Dizaji N,Rahbarghazi R,Garjani A
12-0319-42 was used in Flow cytometry/Cell sorting to highlight the disturbing effects of a diabetic hyperglycemia on either biochemical or biophysical properties of endothelial cells.
Wed Mar 01 00:00:00 UTC 2017
CXCL12/CXCR4 activation by cancer-associated fibroblasts promotes integrin β1 clustering and invasiveness in gastric cancer.
International journal of cancer
Izumi D,Ishimoto T,Miyake K,Sugihara H,Eto K,Sawayama H,Yasuda T,Kiyozumi Y,Kaida T,Kurashige J,Imamura Y,Hiyoshi Y,Iwatsuki M,Iwagami S,Baba Y,Sakamoto Y,Miyamoto Y,Yoshida N,Watanabe M,Takamori H,Araki N,Tan P,Baba H
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Tue Mar 01 00:00:00 UTC 2016
Controlled release hydrogen sulfide delivery system based on mesoporous silica nanoparticles protects graft endothelium from ischemia-reperfusion injury.
International journal of nanomedicine
Wang W,Sun X,Zhang H,Yang C,Liu Y,Yang W,Guo C,Wang C
12-0319 was used in Flow cytometry/Cell sorting to indicate that DATS-MSN, releasing H2S in a controlled release fashion, could serve as an ideal H2S donor.
Mon Mar 06 00:00:00 UTC 2017
Lysyl hydroxylase 2 induces a collagen cross-link switch in tumor stroma.
The Journal of clinical investigation
Chen Y,Terajima M,Yang Y,Sun L,Ahn YH,Pankova D,Puperi DS,Watanabe T,Kim MP,Blackmon SH,Rodriguez J,Liu H,Behrens C,Wistuba II,Minelli R,Scott KL,Sanchez-Adams J,Guilak F,Pati D,Thilaganathan N,Burns AR,Creighton CJ,Martinez ED,Zal T,Grande-Allen KJ,Yamauchi M,Kurie JM
12-0319 was used in Flow cytometry/Cell sorting to demonstrate that epithelial tumourigenesis is accompanied by changes in the biochemical type of collagen cross-links.
Mon Mar 02 00:00:00 UTC 2015
Mesenchymal stromal cells form vascular tubes when placed in fibrin sealant and accelerate wound healing in vivo.
Biomaterials
Mendez JJ,Ghaedi M,Sivarapatna A,Dimitrievska S,Shao Z,Osuji CO,Steinbacher DM,Leffell DJ,Niklason LE
12-0319 was used in Flow cytometry/Cell sorting to show that ADSCs are able to form tubular structures within fibrin gels, and may also contribute to faster wound healing.
Sun Feb 01 00:00:00 UTC 2015
Activation of the Wnt/planar cell polarity pathway is required for pericyte recruitment during pulmonary angiogenesis.
The American journal of pathology
Yuan K,Orcholski ME,Panaroni C,Shuffle EM,Huang NF,Jiang X,Tian W,Vladar EK,Wang L,Nicolls MR,Wu JY,de Jesus Perez VA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Thu Jan 01 00:00:00 UTC 2015
Human esophageal myofibroblasts secrete proinflammatory cytokines in response to acid and Toll-like receptor 4 ligands.
American journal of physiology. Gastrointestinal and liver physiology
Gargus M,Niu C,Vallone JG,Binkley J,Rubin DC,Shaker A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Mon Jun 01 00:00:00 UTC 2015
Human adipose stem cell and ASC-derived cardiac progenitor cellular therapy improves outcomes in a murine model of myocardial infarction.
Stem cells and cloning : advances and applications
Davy PM,Lye KD,Mathews J,Owens JB,Chow AY,Wong L,Moisyadi S,Allsopp RC
12-0319 was used in Flow cytometry/Cell sorting to investigate the potential use of adipose-derived stem cells.
Wed Nov 25 00:00:00 UTC 2015
Vascular progenitors from cord blood-derived induced pluripotent stem cells possess augmented capacity for regenerating ischemic retinal vasculature.
Circulation
Park TS,Bhutto I,Zimmerlin L,Huo JS,Nagaria P,Miller D,Rufaihah AJ,Talbot C,Aguilar J,Grebe R,Merges C,Reijo-Pera R,Feldman RA,Rassool F,Cooke J,Lutty G,Zambidis ET
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Tue Jan 21 00:00:00 UTC 2014
Comparison of human adipose-derived stem cells isolated from subcutaneous, omental, and intrathoracic adipose tissue depots for regenerative applications.
Stem cells translational medicine
Russo V,Yu C,Belliveau P,Hamilton A,Flynn LE
12-0319 was used in Flow cytometry/Cell sorting to determine that depot selection is an important factor to consider when applying ASCs in tissue-specific cell-based regenerative therapies.
Sat Feb 01 00:00:00 UTC 2014
Human mesenchymal stromal cells transiently increase cytokine production by activated T cells before suppressing T-cell proliferation: effect of interferon-γ and tumor necrosis factor-α stimulation.
Cytotherapy
Cuerquis J,Romieu-Mourez R,François M,Routy JP,Young YK,Zhao J,Eliopoulos N
12-0319 was used in Flow cytometry/Cell sorting to assess the kinetic effects of mesenchymal stromal cells on cytokine production and T-cell proliferation, cell cycle and apoptosis.
Sat Feb 01 00:00:00 UTC 2014
Hypoxia-cultured human adipose-derived mesenchymal stem cells are non-oncogenic and have enhanced viability, motility, and tropism to brain cancer.
Cell death & disease
Feng Y,Zhu M,Dangelmajer S,Lee YM,Wijesekera O,Castellanos CX,Denduluri A,Chaichana KL,Li Q,Zhang H,Levchenko A,Guerrero-Cazares H,Quiñones-Hinojosa A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Thu Dec 11 00:00:00 UTC 2014
Efficient and simultaneous generation of hematopoietic and vascular progenitors from human induced pluripotent stem cells.
Cytometry. Part A : the journal of the International Society for Analytical Cytology
Park TS,Zimmerlin L,Zambidis ET
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Tue Jan 01 00:00:00 UTC 2013
Efficient direct reprogramming of mature amniotic cells into endothelial cells by ETS factors and TGFβ suppression.
Cell
Ginsberg M,James D,Ding BS,Nolan D,Geng F,Butler JM,Schachterle W,Pulijaal VR,Mathew S,Chasen ST,Xiang J,Rosenwaks Z,Shido K,Elemento O,Rabbany SY,Rafii S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Fri Oct 26 00:00:00 UTC 2012
Cells expressing the C/EBPbeta isoform, LIP, engulf their neighbors.
PloS one
Abreu M,Sealy L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0319-42) in Flow Cytometry
Thu Apr 11 00:00:00 UTC 2013
IL-7 sustains CD31 expression in human naive CD4+ T cells and preferentially expands the CD31+ subset in a PI3K-dependent manner.
Blood
Azevedo RI,Soares MV,Barata JT,Tendeiro R,Serra-Caetano A,Victorino RM,Sousa AE
12-0319 was used in Flow cytometry/Cell sorting to suggest that during adulthood CD3(+) naive CD4(+) T-cells are maintained by IL-7.
Thu Mar 26 00:00:00 UTC 2009