Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence:
SPLSTPTSA QAAGPSSGSC PPTKFQCRTS GLCVPLTWRC DRDLDCSDGS DEEECRIEPC TQKGQCPPPP GLPCPCTGVS DCSGGTDKKL RNCSRLACLA GELRCTLSDD CIPLTWRCDG HPDCPDSSDE LGCGTNEILP EGDATTMGPP VTLESVTSLR NATTMGPPVT LESVPSVGNA TSSSAGDQSG SPTAY
Germinal center-B (GC-B) cells differentiate into memory B-cells and plasma cells (PC) through interaction with T-cells and follicular dendritic cells (FDC). CD320 augments the proliferation of PC precursors generated by IL-10. Receptor for the cellular uptake of transcobalamin bound cobalamin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 8D6 antigen; 8D6AFDC-SM-8D6; 8D6TCBLR; CD320; CD320 antigen; FDC-signaling molecule 8D6; FDC-SM-8D6; TCblR; Transcobalamin receptor
Gene Aliases: 8D6; 8D6A; CD320; TCBLR; UNQ198/PRO224
UniProt ID: (Human) Q9NPF0
Entrez Gene ID: (Human) 51293
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support