Product References
Immunometabolic interference between cancer and COVID-19.
Frontiers in immunology
Consonni FM,Durante B,Manfredi M,Bleve A,Pandolfo C,Garlatti V,Vanella VV,Marengo E,Barberis E,Bottazzi B,Bombace S,My I,Condorelli G,Torri V,Sica A
Published figure using CD38 monoclonal antibody (Product # 17-0389-42) in Flow Cytometry
Tue Apr 18 00:00:00 UTC 2023
B cell-intrinsic changes with age do not impact antibody-secreting cell formation but delay B cell participation in the germinal centre reaction.
Aging cell
Lee JL,Fra-Bido SC,Burton AR,Innocentin S,Hill DL,Linterman MA
17-0389-42 was used in Flow cytometry/Cell sorting to show that B cells from aged individuals are not intrinsically defective in responding to stimulation and becoming antibody-secreting cells, implicating B cell-extrinsic factors as the primary cause of age-associated impairment in the humoral immunity.
Thu Sep 01 00:00:00 UTC 2022
B cell-intrinsic changes with age do not impact antibody-secreting cell formation but delay B cell participation in the germinal centre reaction.
Aging cell
Lee JL,Fra-Bido SC,Burton AR,Innocentin S,Hill DL,Linterman MA
17-0389-42 was used in Flow cytometry/Cell sorting to show that B cells from aged individuals are not intrinsically defective in responding to stimulation and becoming antibody-secreting cells, implicating B cell-extrinsic factors as the primary cause of age-associated impairment in the humoral immunity.
Thu Sep 01 00:00:00 UTC 2022
Novel myeloma patient-derived xenograft models unveil the potency of anlotinib to overcome bortezomib resistance.
Frontiers in oncology
Yue Y,Cao Y,Mao X,Wang F,Fan P,Qian L,Guo S,Li F,Guo Y,Chen T,Lin Y,Dong W,Liu Y,Huang Y,Gu W
Published figure using CD38 monoclonal antibody (Product # 17-0389-42) in Flow Cytometry
Tue Aug 23 00:00:00 UTC 2022
scTAM-seq enables targeted high-confidence analysis of DNA methylation in single cells.
Genome biology
Bianchi A,Scherer M,Zaurin R,Quililan K,Velten L,Beekman R
17-0389-42 was used in Flow cytometry/Cell sorting to demonstrate that scTAM-seq can resolve DNA methylation dynamics across B-cell differentiation in blood and bone marrow, identifying intermediate differentiation states that were previously masked.
Fri Oct 28 00:00:00 UTC 2022
Kaposi's sarcoma-associated herpesvirus latency-associated nuclear antigen dysregulates expression of MCL-1 by targeting FBW7.
PLoS pathogens
Kim YJ,Kim Y,Kumar A,Kim CW,Toth Z,Cho NH,Lee HR
Published figure using CD38 monoclonal antibody (Product # 17-0389-42) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
Highly efficient CRISPR-Cas9-mediated gene knockout in primary human B cells for functional genetic studies of Epstein-Barr virus infection.
PLoS pathogens
Akidil E,Albanese M,Buschle A,Ruhle A,Pich D,Keppler OT,Hammerschmidt W
Published figure using CD38 monoclonal antibody (Product # 17-0389-42) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2021
SARS-CoV-2 infection paralyzes cytotoxic and metabolic functions of the immune cells.
Heliyon
Singh Y,Trautwein C,Fendel R,Krickeberg N,Berezhnoy G,Bissinger R,Ossowski S,Salker MS,Casadei N,Riess O
Published figure using CD38 monoclonal antibody (Product # 17-0389-42) in Flow Cytometry
Tue Jun 01 00:00:00 UTC 2021
Enhanced CXCR4 Expression of Human CD8Low T Lymphocytes Is Driven by S1P4.
Frontiers in immunology
Burkard T,Dreis C,Herrero San Juan M,Huhn M,Weigert A,Pfeilschifter JM,Radeke HH
17-0389-42 was used in Flow Cytometry to investigate the influence of IL-33 and S1P receptor 4 (S1P4) on the differentiation and migration of human CD8+ T lymphocytes.
Wed Oct 20 00:00:00 UTC 2021
Enhanced CXCR4 Expression of Human CD8Low T Lymphocytes Is Driven by S1P4.
Frontiers in immunology
Burkard T,Dreis C,Herrero San Juan M,Huhn M,Weigert A,Pfeilschifter JM,Radeke HH
17-0389-42 was used in Flow Cytometry to investigate the influence of IL-33 and S1P receptor 4 (S1P4) on the differentiation and migration of human CD8+ T lymphocytes.
Wed Oct 20 00:00:00 UTC 2021
Transitions in lineage specification and gene regulatory networks in hematopoietic stem/progenitor cells over human development.
Cell reports
Roy A,Wang G,Iskander D,O'Byrne S,Elliott N,O'Sullivan J,Buck G,Heuston EF,Wen WX,Meira AR,Hua P,Karadimitris A,Mead AJ,Bodine DM,Roberts I,Psaila B,Thongjuea S
17-0389-42 was used in Flow cytometry/Cell sorting to demonstrate the utility of this dataset for understanding aberrant hematopoiesis through comparison to two cancers that present at distinct time points in postnatal life-juvenile myelomonocytic leukemia, a childhood cancer, and myelofibrosis, which classically presents in older adults.
Tue Sep 14 00:00:00 UTC 2021
Genetic manipulation and immortalized culture of ex vivo primary human germinal center B cells.
Nature protocols
Caeser R,Gao J,Di Re M,Gong C,Hodson DJ
17-0389-42 was used in Flow Cytometry to develop a methodology for the purification and ex vivo expansion of primary, nonmalignant human germinal center B cells.
Sat May 01 00:00:00 UTC 2021
Chronic myelomonocytic leukaemia stem cell transcriptomes anticipate disease morphology and outcome.
EBioMedicine
Wiseman DH,Baker SM,Dongre AV,Gurashi K,Storer JA,Somervaille TC,Batta K
Published figure using CD38 monoclonal antibody (Product # 17-0389-42) in Flow Cytometry
Sat Aug 01 00:00:00 UTC 2020
Patient iPSC-Derived Macrophages to Study Inborn Errors of the IFN-γ Responsive Pathway.
Cells
Haake K,Neehus AL,Buchegger T,Kühnel MP,Blank P,Philipp F,Oleaga-Quintas C,Schulz A,Grimley M,Goethe R,Jonigk D,Kalinke U,Boisson-Dupuis S,Casanova JL,Bustamante J,Lachmann N
17-0389 was used in Flow cytometry/Cell sorting to report a disease modeling platform to study the role of macrophages in patients with inborn errors of IFN-γ immunity.
Wed Feb 19 00:00:00 UTC 2020
IFNγ induces epigenetic programming of human T-bethi B cells and promotes TLR7/8 and IL-21 induced differentiation.
eLife
Zumaquero E,Stone SL,Scharer CD,Jenks SA,Nellore A,Mousseau B,Rosal-Vela A,Botta D,Bradley JE,Wojciechowski W,Ptacek T,Danila MI,Edberg JC,Bridges SL,Kimberly RP,Chatham WW,Schoeb TR,Rosenberg AF,Boss JM,Sanz I,Lund FE
Published figure using CD38 monoclonal antibody (Product # 17-0389-42) in Flow Cytometry
Wed May 15 00:00:00 UTC 2019
A transcriptionally and functionally distinct PD-1+ CD8+ T cell pool with predictive potential in non-small-cell lung cancer treated with PD-1 blockade.
Nature medicine
Thommen DS,Koelzer VH,Herzig P,Roller A,Trefny M,Dimeloe S,Kiialainen A,Hanhart J,Schill C,Hess C,Savic Prince S,Wiese M,Lardinois D,Ho PC,Klein C,Karanikas V,Mertz KD,Schumacher TN,Zippelius A
17-0389 was used in Flow cytometry/Cell sorting to compare transcriptional, metabolic and functional signatures of intratumoral CD8+ T lymphocyte populations with high, intermediate and no PD-1 expression from non-small-cell lung cancer patients.
Sun Jul 01 00:00:00 UTC 2018
Vitamin D supplementation during pregnancy: Effect on the neonatal immune system in a randomized controlled trial.
The Journal of allergy and clinical immunology
Hornsby E,Pfeffer PE,Laranjo N,Cruikshank W,Tuzova M,Litonjua AA,Weiss ST,Carey VJ,O'Connor G,Hawrylowicz C
17-0389 was used in Flow cytometry/Cell sorting to investigate the effect on neonatal immunity of maternal supplementation with 4400 IU/d vitamin D3 during the second and third trimesters of pregnancy by using a subset of cord blood samples from a randomized, double-blind, placebo-controlled clinical trial.
Mon Jan 01 00:00:00 UTC 2018
CD16+NK-92 and anti-CD123 monoclonal antibody prolongs survival in primary human acute myeloid leukemia xenografted mice.
Haematologica
Williams BA,Wang XH,Leyton JV,Maghera S,Deif B,Reilly RM,Minden MD,Keating A
17-0389 was used in Flow cytometry/Cell sorting to study whether CD16+ NK-92 cell line combined with an antibody targeting a leukaemic stem cell antigen can lead to improved survival in a relevant pre-clinical model of acute myeloid leukaemia.
Mon Oct 01 00:00:00 UTC 2018
Genetic Architecture of Adaptive Immune System Identifies Key Immune Regulators.
Cell reports
Lagou V,Garcia-Perez JE,Smets I,Van Horebeek L,Vandebergh M,Chen L,Mallants K,Prezzemolo T,Hilven K,Humblet-Baron S,Moisse M,Van Damme P,Boeckxstaens G,Bowness P,Dubois B,Dooley J,Liston A,Goris A
17-0389-42 was used in Flow Cytometry to identify eight genome-wide significant associations explaining 6%-20% of variance in immune system regulation.
Tue Oct 16 00:00:00 UTC 2018
Abnormal phenotypic features of IgM+B cell subsets in patients with chronic hepatitis C virus infection.
Experimental and therapeutic medicine
Kong F,Feng B,Zhang H,Rao H,Wang J,Cong X,Wei L
Tue Aug 01 00:00:00 UTC 2017
Effects of acute exposure to low-dose radiation on the characteristics of human bone marrow mesenchymal stromal/stem cells.
Inflammation and regeneration
Fujishiro A,Miura Y,Iwasa M,Fujii S,Sugino N,Andoh A,Hirai H,Maekawa T,Ichinohe T
17-0389 was used in Flow cytometry/Cell sorting to determine whether low dose irradiation affects the function of mesenchymal stromal/stem cells.
Fri Mar 11 00:00:00 UTC 2022
PD-L1 mediated the differentiation of tumor-infiltrating CD19+ B lymphocytes and T cells in Invasive breast cancer.
Oncoimmunology
Guan H,Lan Y,Wan Y,Wang Q,Wang C,Xu L,Chen Y,Liu W,Zhang X,Li Y,Gu Y,Wang Z,Xie F
17-0389 was used in Flow cytometry/Cell sorting to investigate the role of B cells in human breast cancer etiology.
Mon Feb 01 00:00:00 UTC 2016
Renal Transplant Recipients Treated with Calcineurin-Inhibitors Lack Circulating Immature Transitional CD19+CD24hiCD38hi Regulatory B-Lymphocytes.
PloS one
Tebbe B,Wilde B,Ye Z,Wang J,Wang X,Jian F,Dolff S,Schedlowski M,Hoyer PF,Kribben A,Witzke O,Hoerning A
17-0389 was used in Flow cytometry/Cell sorting to analyse the effect of calcineurin inhibition on peripherally circulating regulatory B cells in renal transplant recipients receiving cyclosporine A or tacrolimus.
Mon Aug 29 00:00:00 UTC 2016
SF3B1 mutant MDS-initiating cells may arise from the haematopoietic stem cell compartment.
Nature communications
Mian SA,Rouault-Pierre K,Smith AE,Seidl T,Pizzitola I,Kizilors A,Kulasekararaj AG,Bonnet D,Mufti GJ
17-0389 was used in Flow cytometry/Cell sorting to demonstrate that mutations to SF3B1 can arise from CD34(+)CD38(-)CD45RA(-)CD90(+)CD49f(+) haematopoietic stem cells and is an initiating event in myelodysplastic syndrome patients.
Tue Dec 08 00:00:00 UTC 2015
Immunologic human renal allograft injury associates with an altered IL-10/TNF-α expression ratio in regulatory B cells.
Journal of the American Society of Nephrology : JASN
Cherukuri A,Rothstein DM,Clark B,Carter CR,Davison A,Hernandez-Fuentes M,Hewitt E,Salama AD,Baker RJ
17-0389 was used in Flow cytometry/Cell sorting to explore proinflammatory cytokine expression in B-cell subsets.
Tue Jul 01 00:00:00 UTC 2014
Generation of functional, antigen-specific CD8+ human T cells from cord blood stem cells using exogenous Notch and tetramer-TCR signaling.
Stem cells (Dayton, Ohio)
Fernandez I,Ooi TP,Roy K
Published figure using CD38 monoclonal antibody (Product # 17-0389-42) in Flow Cytometry
Wed Jan 01 00:00:00 UTC 2014
Generation of functional, antigen-specific CD8+ human T cells from cord blood stem cells using exogenous Notch and tetramer-TCR signaling.
Stem cells (Dayton, Ohio)
Fernandez I,Ooi TP,Roy K
Published figure using CD38 monoclonal antibody (Product # 17-0389-42) in Flow Cytometry
Wed Jan 01 00:00:00 UTC 2014
DNA methylation profiling in human B cells reveals immune regulatory elements and epigenetic plasticity at Alu elements during B-cell activation.
Genome research
Lai AY,Mav D,Shah R,Grimm SA,Phadke D,Hatzi K,Melnick A,Geigerman C,Sobol SE,Jaye DL,Wade PA
17-0389 was used in Flow cytometry/Cell sorting to investigate the profile of B-lymphocyte subsets during different stages of the humoral immune response in humans.
Sun Dec 01 00:00:00 UTC 2013
Targeting CDK1 promotes FLT3-activated acute myeloid leukemia differentiation through C/EBPα.
The Journal of clinical investigation
Radomska HS,Alberich-Jordà M,Will B,Gonzalez D,Delwel R,Tenen DG
Published figure using CD38 monoclonal antibody (Product # 17-0389-42) in Flow Cytometry
Wed Aug 01 00:00:00 UTC 2012
Human tonsil B-cell lymphoma 6 (BCL6)-expressing CD4+ T-cell subset specialized for B-cell help outside germinal centers.
Proceedings of the National Academy of Sciences of the United States of America
Bentebibel SE,Schmitt N,Banchereau J,Ueno H
17-0389 was used in Flow cytometry/Cell sorting to identify and investigate the role of a previously undefined BCL6-expressing CD4(+) T-cell subset in human tonsils.
Tue Aug 16 00:00:00 UTC 2011
Further phenotypic characterization of the primitive lineage- CD34+CD38-CD90+CD45RA- hematopoietic stem cell/progenitor cell sub-population isolated from cord blood, mobilized peripheral blood and patients with chronic myelogenous leukemia.
Blood cancer journal
Wisniewski D,Affer M,Willshire J,Clarkson B
17-0389 was used in Flow cytometry/Cell sorting to investigate the purification of haematopoietic stem cells, and the potential targetable differences between primitive normal and chronic myelogenous leukaemia stem/progenitor cells.
Thu Sep 01 00:00:00 UTC 2011
Fc receptor-like 3 protein expressed on IL-2 nonresponsive subset of human regulatory T cells.
Journal of immunology (Baltimore, Md. : 1950)
Nagata S,Ise T,Pastan I
17-0389 was used in Flow cytometry/Cell sorting to study the effects of Foxp3 and IL-2 on FCRL3-/+ nTreg cell subsets.
Mon Jun 15 00:00:00 UTC 2009