Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • CD3 Antibodies

            Invitrogen

            CD3e Monoclonal Antibody (145-2C11), APC, eBioscience™

            Advanced Verification
            97 Published Figures
            230 References
            View all (543) CD3 antibodies

            Promotions

            View available promotion(s)

            Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # 17-0031-82
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            271.15
            Special offer
            Online exclusive
            319.00
            Save 47.85 (15%)
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite CD3e Monoclonal Antibody (145-2C11), APC, eBioscience™

            • Antibody Testing Data (1)
            • Published Figures (97)
            • Advanced Verification (1)
            Application
            CD3e Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 99

            CD3e Antibody (17-0031-82) in Flow

            Staining of C57Bl/6 splenocytes with Anti-Human/Mouse CD45R (B220) FITC (Product # 11-0452-82) and 0.125 µg of Armenian Hamster IgG Isotype Control APC (Product # 17-4888-82) (left) or 0.125 µg of Anti-Mouse CD3e APC (right). Total viable cells were used for analysis.
            View Product

            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody in Flow Cytometry (Flow)
            CD3e Antibody

            Product Details

            17-0031-82

            Applications
            Tested Dilution
            Publications

            Immunohistochemistry (IHC)

            -
            View 6 publications 6 publications

            Immunohistochemistry (Frozen) (IHC (F))

            -
            View 2 publications 2 publications

            Flow Cytometry (Flow)

            0.25 µg/test
            View 219 publications 219 publications

            Functional Assay (Functional)

            -
            View 2 publications 2 publications

            Miscellaneous PubMed (Misc)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Mouse

            Published species

            Human, Mouse

            Host/Isotype

            Armenian hamster / IgG

            Recommended Isotype Control

            Armenian Hamster IgG Isotype Control (eBio299Arm), APC, eBioscience™

            Class

            Monoclonal

            Type

            Antibody

            Clone

            145-2C11

            Conjugate

            APC
            • Unconjugated
            • Alexa Fluor 488 
            • APC-eFluor 780
            • Biotin 
            • Brilliant UV 496
            • Brilliant UV 563
            • Brilliant UV 615
            • Brilliant UV 661
            • Brilliant UV 737
            • Brilliant Violet 421
            • Brilliant Violet 480
            • Brilliant Violet 605
            • Brilliant Violet 650
            • Brilliant Violet 711
            • Brilliant Violet 786
            • eFluor 450
            • FITC
            • Functional Grade
            • NovaFluor Blue 510
            • NovaFluor Blue 610-70S
            • NovaFluor Blue 660-120S
            • NovaFluor Red 685
            • NovaFluor Red 700
            • NovaFluor Red 710
            • NovaFluor Yellow 610
            • NovaFluor Yellow 660
            • NovaFluor Yellow 690
            • NovaFluor Yellow 700
            • NovaFluor Yellow 730
            • PE 
            • PE-Cyanine5
            • PE-Cyanine5.5
            • PE-Cyanine7
            • PE-eFluor 610
            • PerCP-Cyanine5.5
            • Super Bright 436
            • Super Bright 600
            • Super Bright 780
            • Request custom conjugation

            Excitation/Emission Max

            651/660 nm View spectra spectra

            Form

            Liquid

            Concentration

            0.2 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2

            Contains

            0.09% sodium azide

            Storage conditions

            4°C, store in dark, DO NOT FREEZE!

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_469315

            Product Specific Information

            Description: The 145-2C11 monoclonal antibody reacts with mouse CD3e, a 20 kDa subunit of the TCR complex. Along with the other CD3 subunits, gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Binding of 145-2C11 to TCR initiates the intracellular biochemical pathway resulting in cellular activation, proliferation, and apoptosis depending on specific conditions utilized. 145-2C11 is commonly used as a phenotypic marker for mouse T cells.

            Applications Reported: The 145-2C11 antibody has been reported for use in flow cytometric analysis.

            Applications Tested: The 145-2C11 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

            Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            The CD3 complex, composed of gamma, delta, epsilon, and zeta subunits, is essential for the assembly, trafficking, and surface expression of the T cell receptor (TCR) complex. These subunits are structurally related members of the immunoglobulin superfamily and are encoded by closely linked genes on human chromosome 11. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells, but not on B or NK cells. The CD3 subunits play a crucial role in transducing antigen-recognition signals into the cytoplasm of T cells. The cytoplasmic tails of CD3 subunits contain a double tyrosine-based motif that associates with cytoplasmic signal transduction molecules, mediating T cell activation through the TCR. Crosslinking of the TCR initiates intracellular biochemical pathways that result in cellular activation, proliferation, and potentially growth arrest and cell survival. CD3 is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes, and Purkinje cells in the cerebellum. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. Defects in the CD3 gene are associated with CD3 immunodeficiency, highlighting its importance in immune function and regulation.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            How to build a flow cytometry experiment with Invitrogen Flow Cytometry Panel Builder
            The Invitrogen Flow Cytometry Panel Builder webtool helps you chose antibody clones, fluorophores, fluorescent proteins, and reagents to design your experiment.
            Video Player is loading.
            Current Time 0:00
            /
            Duration 8:21
            Loaded: 0%
            0:00
            Stream Type LIVE
            Remaining Time -8:21
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected
            • en (Main), selected

            This is a modal window.

            Beginning of dialog window. Escape will cancel and close the window.

            Text
            Text Background
            Caption Area Background
            Font Size
            Text Edge Style
            Font Family

            End of dialog window.

            This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

            How to use the Panel Builder

            Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

            Related Products for

            Flow Cytometry
            • Additional Formats
            • Isotype Controls
            • Buffers FC
            • Viability Dyes FC
            • Compensation Beads FC
            • Labeling Kits FC

            Bioinformatics

            Protein Aliases: CD247; CD247 antigen; CD3 antigen, zeta polypeptide; CD3 TCR complex; CD3-zeta/eta; CD3d; CD3e; CD3g; FLJ18683; Leu-4; T cell antigen receptor complex epsilon subunit of T3; T-cell receptor T3 delta chain; T-cell receptor T3 eta chain; T-cell receptor T3 gamma chain; T-cell receptor T3 zeta chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; T-cell surface glycoprotein CD3 zeta chain; T3/TCR complex

            View more View less

            Gene Aliases: 4930549J05Rik; A430104F18Rik; AI504783; AW552088; Cd247; CD3; Cd3-eta; Cd3-zeta; Cd3d; Cd3e; CD3epsilon; Cd3g; Cd3h; Cd3z; Cd3zeta; Ctg-3; Ctg3; T3d; T3e; T3g; T3z; Tcrk; Tcrz

            View more View less

            UniProt ID: (Mouse) P24161, (Mouse) P04235, (Mouse) P22646, (Mouse) P11942

            View more View less

            Entrez Gene ID: (Mouse) 12503, (Mouse) 12500, (Mouse) 12501, (Mouse) 12502

            View more View less

            Function(s)

            transmembrane signaling receptor activity protein binding identical protein binding protein heterodimerization activity signal transducer activity SH3 domain binding defense/immunity protein immunoglobulin receptor superfamily

            Process(es)

            cell surface receptor signaling pathway T cell receptor signaling pathway protein complex assembly T cell activation positive thymic T cell selection positive regulation of T cell anergy response to nutrient T cell costimulation positive regulation of interferon-gamma production positive regulation of interleukin-4 production positive regulation of T cell proliferation negative thymic T cell selection positive regulation of interleukin-2 biosynthetic process negative regulation of smoothened signaling pathway positive regulation of alpha-beta T cell proliferation lymphocyte activation positive regulation of peptidyl-tyrosine phosphorylation positive regulation of calcium-mediated signaling positive regulation of T cell activation apoptotic signaling pathway establishment or maintenance of cell polarity protein transport regulation of lymphocyte apoptotic process
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-4dj9n:80/100.66.77.185:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0