Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • CD4 Antibodies

            Invitrogen

            CD4 Monoclonal Antibody (GK1.5), FITC, eBioscience™

            Advanced Verification
            97 Published Figures
            427 References
            View all (661) CD4 antibodies

            Promotions

            View available promotion(s)

            Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # 11-0041-82
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            88.40
            Special offer
            Online exclusive
            104.00
            Save 15.60 (15%)
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite CD4 Monoclonal Antibody (GK1.5), FITC, eBioscience™

            • Antibody Testing Data (1)
            • Published Figures (97)
            • Advanced Verification (1)
            Application
            CD4 Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 99

            CD4 Antibody (11-0041-82) in Flow

            Staining of mouse thymocytes with Anti-Mouse CD8a PE (Product # 12-0081-82) (left) or mouse splenocytes with Anti-Mouse CD3e PE (Product # 12-0031-82) (right) and Anti-Mouse CD4 FITC. Total viable cells were used for analysis.
            View Product

            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody in Flow Cytometry (Flow)
            CD4 Antibody

            Product Details

            11-0041-82

            Applications
            Tested Dilution
            Publications

            Immunohistochemistry (IHC)

            -
            View 21 publications 21 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            -
            View 1 publication 1 publication

            Immunohistochemistry (Frozen) (IHC (F))

            -
            View 2 publications 2 publications

            Immunocytochemistry (ICC/IF)

            -
            View 7 publications 7 publications

            Flow Cytometry (Flow)

            0.25 µg/test
            View 391 publications 391 publications

            Functional Assay (Functional)

            -
            View 2 publications 2 publications

            Miscellaneous PubMed (Misc)

            -
            View 3 publications 3 publications
            Product Specifications

            Species Reactivity

            Mouse

            Published species

            Fish, Human, Mouse, Rat

            Host/Isotype

            Rat / IgG2b, kappa

            Recommended Isotype Control

            Rat IgG2b kappa Isotype Control (eB149/10H5), FITC, eBioscience™

            Class

            Monoclonal

            Type

            Antibody

            Clone

            GK1.5

            Conjugate

            FITC
            • Unconjugated
            • Alexa Fluor 488 
            • Alexa Fluor 532 
            • Alexa Fluor 700 
            • APC 
            • APC-eFluor 780
            • Biotin 
            • Brilliant UV 395
            • Brilliant UV 496
            • Brilliant UV 563
            • Brilliant UV 661
            • Brilliant UV 737
            • Brilliant Violet 480
            • Brilliant Violet 605
            • eFluor 450
            • eFluor 506
            • eFluor 660
            • Functional Grade
            • NovaFluor Blue 510
            • NovaFluor Blue 530
            • NovaFluor Blue 555
            • NovaFluor Blue 585
            • NovaFluor Blue 610-30S
            • NovaFluor Blue 610-70S
            • NovaFluor Blue 660-120S
            • NovaFluor Blue 660-40S
            • NovaFluor Blue 690
            • NovaFluor Blue 725
            • NovaFluor Blue 760
            • NovaFluor Blue 800
            • NovaFluor Red 660
            • NovaFluor Red 685
            • NovaFluor Red 700
            • NovaFluor Red 710
            • NovaFluor UV 765
            • NovaFluor Violet 690
            • NovaFluor Violet 745
            • NovaFluor Violet 800
            • NovaFluor Yellow 570
            • NovaFluor Yellow 590
            • NovaFluor Yellow 610
            • NovaFluor Yellow 660
            • NovaFluor Yellow 690
            • NovaFluor Yellow 700
            • NovaFluor Yellow 730
            • NovaFluor Yellow 810
            • PE 
            • PE-Cyanine5
            • PE-Cyanine7
            • PE-eFluor 610
            • PerCP-eFluor 710
            • Super Bright 436
            • Super Bright 600
            • Super Bright 645
            • Super Bright 702
            • Super Bright 780
            • Request custom conjugation

            Excitation/Emission Max

            498/517 nm View spectra spectra

            Form

            Liquid

            Concentration

            0.5 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2

            Contains

            0.09% sodium azide

            Storage conditions

            4°C, store in dark, DO NOT FREEZE!

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_464892

            Product Specific Information

            Description: The GK1.5 monoclonal antibody reacts with the mouse CD4 molecule, a 55 kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of GK1.5 is blocked by RM4-5.

            Applications Reported: The GK1.5 antibody has been reported for use in flow cytometric analysis.

            Applications Tested: This GK1.5 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.25 µg per test A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

            Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            How to build a flow cytometry experiment with Invitrogen Flow Cytometry Panel Builder
            The Invitrogen Flow Cytometry Panel Builder webtool helps you chose antibody clones, fluorophores, fluorescent proteins, and reagents to design your experiment.
            Video Player is loading.
            Current Time 0:00
            /
            Duration 0:00
            Loaded: 0%
            0:00
            Stream Type LIVE
            Remaining Time -0:00
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected
            • en (Main), selected

            This is a modal window.

            Beginning of dialog window. Escape will cancel and close the window.

            Text
            Text Background
            Caption Area Background
            Font Size
            Text Edge Style
            Font Family

            End of dialog window.

            This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

            How to use the Panel Builder

            Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

            Related Products for

            Flow Cytometry
            • Additional Formats
            • Isotype Controls
            • Buffers FC
            • Viability Dyes FC
            • Compensation Beads FC
            • Labeling Kits FC
            Catalog #
            14-0041-82
            Invitrogen
            CD4 Monoclonal Antibody (GK1.5), eBioscience™
            Unit Size :
            100 µg
            Catalog #
            13-0041-82
            Invitrogen
            CD4 Monoclonal Antibody (GK1.5), Biotin, eBioscience™
            Unit Size :
            100 µg
            Catalog #
            16-0041-82
            Invitrogen
            CD4 Monoclonal Antibody (GK1.5), Functional Grade, eBioscience™
            Unit Size :
            100 µg
            Catalog #
            78-0041-82
            Invitrogen
            CD4 Monoclonal Antibody (GK1.5), Super Bright™ 780, eBioscience™
            Unit Size :
            100 µg
            Catalog #
            62-0041-82
            Invitrogen
            CD4 Monoclonal Antibody (GK1.5), Super Bright™ 436, eBioscience™
            Unit Size :
            100 µg
            Catalog #
            M001T02Y05-A
            Invitrogen
            CD4 Monoclonal Antibody (GK1.5), NovaFluor™ Yellow 690, eBioscience™
            Unit Size :
            25 µg
            Catalog #
            376-0041-82
            Invitrogen
            CD4 Monoclonal Antibody (GK1.5), Brilliant Ultra Violet™ 661, eBioscience™
            Unit Size :
            100 µg
            Catalog #
            365-0041-82
            Invitrogen
            CD4 Monoclonal Antibody (GK1.5), Brilliant Ultra Violet™ 563, eBioscience™
            Unit Size :
            100 µg
            Catalog #
            58-0041-82
            Invitrogen
            CD4 Monoclonal Antibody (GK1.5), Alexa Fluor™ 532, eBioscience™
            Unit Size :
            100 µg
            Catalog #
            406-0041-82
            Invitrogen
            CD4 Monoclonal Antibody (GK1.5), Brilliant Violet™ 605, eBioscience™
            Unit Size :
            100 µg

            Bioinformatics

            Protein Aliases: CD4; CD4 antigen p55; cd4a; fCD4; Leu-3; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4)

            View more View less

            Gene Aliases: Cd4; L3T4; Ly-4

            View more View less

            UniProt ID: (Mouse) P06332

            View more View less

            Entrez Gene ID: (Mouse) 12504

            View more View less

            Function(s)

            glycoprotein binding protein binding zinc ion binding coreceptor activity enzyme binding protein kinase binding protein homodimerization activity

            Process(es)

            cytokine production immune system process induction by virus of host cell-cell fusion immune response cell adhesion cell surface receptor signaling pathway T cell differentiation maintenance of protein location in cell T cell activation T cell selection protein palmitoleylation positive regulation of protein kinase activity positive regulation of peptidyl-tyrosine phosphorylation defense response to Gram-negative bacterium positive regulation of calcium-mediated signaling regulation of T cell activation positive regulation of T cell activation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-wfwm6:80/100.66.76.90:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0