Product References
Schlafen14 Impairs HIV-1 Expression in a Codon Usage-Dependent Manner.
Viruses
Valenzuela C,Saucedo S,Llano M
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Mon Mar 25 00:00:00 UTC 2024
Endosialin-positive tumor-derived pericytes promote tumor progression through impeding the infiltration of CD8+ T cells in clear cell renal cell carcinoma.
Cancer immunology, immunotherapy : CII
Lu T,Zhang J,Lu S,Yang F,Gan L,Wu X,Song H,Liu S,Xu C,Han D,Yang B,Wen W,Qin W,Yang L
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Thu Jun 01 00:00:00 UTC 2023
Distinct SARS-CoV-2 specific NLRP3 and IL-1β responses in T cells of aging patients during acute COVID-19 infection.
Frontiers in immunology
Mahalingam SS,Jayaraman S,Arunkumar A,Dudley HM,Anthony DD,Shive CL,Jacobson JM,Pandiyan P
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Thu Oct 12 00:00:00 UTC 2023
Estrogen Protects against Renal Ischemia-Reperfusion Injury by Regulating Th17/Treg Cell Immune Balance.
Disease markers
Zhang Y,Chang Y,Han Z,Ma K,Zeng X,Li L
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Tue Oct 18 00:00:00 UTC 2022
Plasma Extracellular Vesicle Subtypes May be Useful as Potential Biomarkers of Immune Activation in People With HIV.
Pathogens & immunity
Bazié WW,Boucher J,Vitry J,Goyer B,Routy JP,Tremblay C,Trottier S,Jenabian MA,Provost P,Alary M,Gilbert C
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Sat Apr 23 00:00:00 UTC 2022
Targeting Phosphatidylserine Enhances the Anti-tumor Response to Tumor-Directed Radiation Therapy in a Preclinical Model of Melanoma.
Cell reports
Budhu S,Giese R,Gupta A,Fitzgerald K,Zappasodi R,Schad S,Hirschhorn D,Campesato LF,De Henau O,Gigoux M,Liu C,Mazo G,Deng L,Barker CA,Wolchok JD,Merghoub T
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Tue Jan 12 00:00:00 UTC 2021
Small-molecule antagonist of VLA-4 (GW559090) attenuated neuro-inflammation by targeting Th17 cell trafficking across the blood-retinal barrier in experimental autoimmune uveitis.
Journal of neuroinflammation
Chen YH,Eskandarpour M,Zhang X,Galatowicz G,Greenwood J,Lightman S,Calder V
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Thu Feb 18 00:00:00 UTC 2021
A Cross-Reactive Small Protein Binding Domain Provides a Model to Study Off-Tumor CAR-T Cell Toxicity.
Molecular therapy oncolytics
Hammill JA,Kwiecien JM,Dvorkin-Gheva A,Lau VWC,Baker C,Wu Y,Bezverbnaya K,Aarts C,Heslen CW,Denisova GF,Derocher H,Milne K,Nelson BH,Bramson JL
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Fri Jun 26 00:00:00 UTC 2020
The Potential of Nanobody-Targeted Photodynamic Therapy to Trigger Immune Responses.
Cancers
Beltrán Hernández I,Angelier ML,Del Buono D'Ondes T,Di Maggio A,Yu Y,Oliveira S
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Wed Apr 15 00:00:00 UTC 2020
T-Lymphocyte Subset Distribution and Activity in Patients With Glaucoma.
Investigative ophthalmology & visual science
Yang X,Zeng Q,Göktas E,Gopal K,Al-Aswad L,Blumberg DM,Cioffi GA,Liebmann JM,Tezel G
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2019
Umbilical cord blood‑derived Helios‑positive regulatory T cells promote angiogenesis in acute lymphoblastic leukemia in mice via CCL22 and the VEGFA‑VEGFR2 pathway.
Molecular medicine reports
Li X,Li D,Shi Q,Huang X,Ju X
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Wed May 01 00:00:00 UTC 2019
Intracellular delivery of mRNA to human primary T cells with microfluidic vortex shedding.
Scientific reports
Jarrell JA,Twite AA,Lau KHWJ,Kashani MN,Lievano AA,Acevedo J,Priest C,Nieva J,Gottlieb D,Pawell RS
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2019
Expression of CD20 after viral reactivation renders HIV-reservoir cells susceptible to Rituximab.
Nature communications
Serra-Peinado C,Grau-Expósito J,Luque-Ballesteros L,Astorga-Gamaza A,Navarro J,Gallego-Rodriguez J,Martin M,Curran A,Burgos J,Ribera E,Raventós B,Willekens R,Torrella A,Planas B,Badía R,Garcia F,Castellví J,Genescà M,Falcó V,Buzon MJ
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Fri Aug 16 00:00:00 UTC 2019
Accumulation of T-helper 22 cells, interleukin-22 and myeloid-derived suppressor cells promotes gastric cancer progression in elderly patients.
Oncology letters
Chen X,Wang Y,Wang J,Wen J,Jia X,Wang X,Zhang H
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Sun Jul 01 00:00:00 UTC 2018
IL-6 receptor blockade corrects defects of XIAP-deficient regulatory T cells.
Nature communications
Hsieh WC,Hsu TS,Chang YJ,Lai MZ
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Wed Jan 31 00:00:00 UTC 2018
Complement receptor CD46 co-stimulates optimal human CD8+ T cell effector function via fatty acid metabolism.
Nature communications
Arbore G,West EE,Rahman J,Le Friec G,Niyonzima N,Pirooznia M,Tunc I,Pavlidis P,Powell N,Li Y,Liu P,Servais A,Couzi L,Fremeaux-Bacchi V,Placais L,Ferraro A,Walsh PR,Kavanagh D,Afzali B,Lavender P,Lachmann HJ,Kemper C
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Wed Oct 10 00:00:00 UTC 2018
Heterogeneous Tumor-Immune Microenvironments among Differentially Growing Metastases in an Ovarian Cancer Patient.
Cell
Jiménez-Sánchez A,Memon D,Pourpe S,Veeraraghavan H,Li Y,Vargas HA,Gill MB,Park KJ,Zivanovic O,Konner J,Ricca J,Zamarin D,Walther T,Aghajanian C,Wolchok JD,Sala E,Merghoub T,Snyder A,Miller ML
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Thu Aug 24 00:00:00 UTC 2017
Ex vivo culture of human atherosclerotic plaques: A model to study immune cells in atherogenesis.
Atherosclerosis
Lebedeva A,Vorobyeva D,Vagida M,Ivanova O,Felker E,Fitzgerald W,Danilova N,Gontarenko V,Shpektor A,Vasilieva E,Margolis L
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Fri Dec 01 00:00:00 UTC 2017
Hormonal vitamin D up-regulates tissue-specific PD-L1 and PD-L2 surface glycoprotein expression in humans but not mice.
The Journal of biological chemistry
Dimitrov V,Bouttier M,Boukhaled G,Salehi-Tabar R,Avramescu RG,Memari B,Hasaj B,Lukacs GL,Krawczyk CM,White JH
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Fri Dec 15 00:00:00 UTC 2017
Targeted Disruption of TCF12 Reveals HEB as Essential in Human Mesodermal Specification and Hematopoiesis.
Stem cell reports
Li Y,Brauer PM,Singh J,Xhiku S,Yoganathan K,Zúñiga-Pflücker JC,Anderson MK
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Tue Sep 12 00:00:00 UTC 2017
Single-cell profiling reveals GPCR heterogeneity and functional patterning during neuroinflammation.
JCI insight
Tischner D,Grimm M,Kaur H,Staudenraus D,Carvalho J,Looso M,Günther S,Wanke F,Moos S,Siller N,Breuer J,Schwab N,Zipp F,Waisman A,Kurschus FC,Offermanns S,Wettschureck N
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Thu Aug 03 00:00:00 UTC 2017
PD-L1 expression and prognostic impact in glioblastoma.
Neuro-oncology
Nduom EK,Wei J,Yaghi NK,Huang N,Kong LY,Gabrusiewicz K,Ling X,Zhou S,Ivan C,Chen JQ,Burks JK,Fuller GN,Calin GA,Conrad CA,Creasy C,Ritthipichai K,Radvanyi L,Heimberger AB
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2016
Altered expression of miR-92a correlates with Th17 cell frequency in patients with primary biliary cirrhosis.
International journal of molecular medicine
Liang DY,Hou YQ,Luo LJ,Ao L
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Fri Jul 01 00:00:00 UTC 2016
Tax and Semaphorin 4D Released from Lymphocytes Infected with Human Lymphotropic Virus Type 1 and Their Effect on Neurite Growth.
AIDS research and human retroviruses
Quintremil S,Alberti C,Rivera M,Medina F,Puente J,Cartier L,Ramírez E,Tanaka Y,Valenzuela MA
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2016
Tumor-derived exosomes regulate expression of immune function-related genes in human T cell subsets.
Scientific reports
Muller L,Mitsuhashi M,Simms P,Gooding WE,Whiteside TL
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Thu Feb 04 00:00:00 UTC 2016
Small CD4 Mimetics Prevent HIV-1 Uninfected Bystander CD4 + T Cell Killing Mediated by Antibody-dependent Cell-mediated Cytotoxicity.
EBioMedicine
Richard J,Veillette M,Ding S,Zoubchenok D,Alsahafi N,Coutu M,Brassard N,Park J,Courter JR,Melillo B,Smith AB,Shaw GM,Hahn BH,Sodroski J,Kaufmann DE,Finzi A
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2016
The HIV-1 gp120 CD4-bound conformation is preferentially targeted by antibody-dependent cellular cytotoxicity-mediating antibodies in sera from HIV-1-infected individuals.
Journal of virology
Veillette M,Coutu M,Richard J,Batraville LA,Dagher O,Bernard N,Tremblay C,Kaufmann DE,Roger M,Finzi A
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Thu Jan 01 00:00:00 UTC 2015
Aging and cytomegalovirus infection differentially and jointly affect distinct circulating T cell subsets in humans.
Journal of immunology (Baltimore, Md. : 1950)
Wertheimer AM,Bennett MS,Park B,Uhrlaub JL,Martinez C,Pulko V,Currier NL,Nikolich-Žugich D,Kaye J,Nikolich-Žugich J
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Sat Mar 01 00:00:00 UTC 2014
An increased abundance of tumor-infiltrating regulatory T cells is correlated with the progression and prognosis of pancreatic ductal adenocarcinoma.
PloS one
Tang Y,Xu X,Guo S,Zhang C,Tang Y,Tian Y,Ni B,Lu B,Wang H
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Immunohistochemistry
Tue Jan 06 00:00:00 UTC 2015
An increased abundance of tumor-infiltrating regulatory T cells is correlated with the progression and prognosis of pancreatic ductal adenocarcinoma.
PloS one
Tang Y,Xu X,Guo S,Zhang C,Tang Y,Tian Y,Ni B,Lu B,Wang H
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Immunohistochemistry
Tue Jan 06 00:00:00 UTC 2015
CD4-CCR5 interaction in intracellular compartments contributes to receptor expression at the cell surface.
Blood
Achour L,Scott MG,Shirvani H,Thuret A,Bismuth G,Labbé-Jullié C,Marullo S
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Thu Feb 26 00:00:00 UTC 2009
CD4-CCR5 interaction in intracellular compartments contributes to receptor expression at the cell surface.
Blood
Achour L,Scott MG,Shirvani H,Thuret A,Bismuth G,Labbé-Jullié C,Marullo S
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Thu Feb 26 00:00:00 UTC 2009
CD4-CCR5 interaction in intracellular compartments contributes to receptor expression at the cell surface.
Blood
Achour L,Scott MG,Shirvani H,Thuret A,Bismuth G,Labbé-Jullié C,Marullo S
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Thu Feb 26 00:00:00 UTC 2009
CD4-CCR5 interaction in intracellular compartments contributes to receptor expression at the cell surface.
Blood
Achour L,Scott MG,Shirvani H,Thuret A,Bismuth G,Labbé-Jullié C,Marullo S
Published figure using CD4 monoclonal antibody (Product # 16-0048-81) in Flow Cytometry
Thu Feb 26 00:00:00 UTC 2009