Product References
DNAJA3 regulates B cell development and immune function.
Biomedical journal
Sayson SL,Fan JN,Ku CL,Lo JF,Chou SH
Published figure using CD43 monoclonal antibody (Product # 11-0431-82) in Flow Cytometry
Mon Apr 01 00:00:00 UTC 2024
A Hyper-IgM Syndrome Mutation in Activation-Induced Cytidine Deaminase Disrupts G-Quadruplex Binding and Genome-wide Chromatin Localization.
Immunity
Yewdell WT,Kim Y,Chowdhury P,Lau CM,Smolkin RM,Belcheva KT,Fernandez KC,Cols M,Yen WF,Vaidyanathan B,Angeletti D,McDermott AB,Yewdell JW,Sun JC,Chaudhuri J
11-0431-82 was used in Flow cytometry/Cell sorting to indicate a crucial role for G4 binding in AID targeting and suggest that AID activity may extend beyond Ig loci to regulate the expression of genes relevant to the physiology and pathology of activated B cells.
Tue Nov 17 00:00:00 UTC 2020
The evolution of zebrafish RAG2 protein is required for adapting to the elevated body temperature of the higher endothermic vertebrates.
Scientific reports
Sun A,Xu K,Liu H,Li H,Shi Y,Zhu X,Liang T,Li X,Cao X,Ji Y,Jiang T,Xu C,Liu X
Published figure using CD43 monoclonal antibody (Product # 11-0431-82) in Flow Cytometry
Thu Mar 05 00:00:00 UTC 2020
Modest Declines in Proteome Quality Impair Hematopoietic Stem Cell Self-Renewal.
Cell reports
Hidalgo San Jose L,Sunshine MJ,Dillingham CH,Chua BA,Kruta M,Hong Y,Hatters DM,Signer RAJ
11-0431 was used in Flow cytometry/Cell sorting to show that hematopoietic stem cells (HSCs) are dependent on low protein synthesis to maintain proteostasis, which promotes their self-renewal.
Tue Jan 07 00:00:00 UTC 2020
Polyamines Control eIF5A Hypusination, TFEB Translation, and Autophagy to Reverse B Cell Senescence.
Molecular cell
Zhang H,Alsaleh G,Feltham J,Sun Y,Napolitano G,Riffelmacher T,Charles P,Frau L,Hublitz P,Yu Z,Mohammed S,Ballabio A,Balabanov S,Mellor J,Simon AK
11-0431 was used in Flow cytometry/Cell sorting to reveal an unexpected autophagy regulatory mechanism mediated by eIF5A at the translational level, which can be harnessed to reverse immune senescence in humans.
Thu Oct 03 00:00:00 UTC 2019
Mutations in topoisomerase IIβ result in a B cell immunodeficiency.
Nature communications
Broderick L,Yost S,Li D,McGeough MD,Booshehri LM,Guaderrama M,Brydges SD,Kucharova K,Patel NC,Harr M,Hakonarson H,Zackai E,Cowell IG,Austin CA,Hügle B,Gebauer C,Zhang J,Xu X,Wang J,Croker BA,Frazer KA,Putnam CD,Hoffman HM
11-0431-82 was used in Flow Cytometry to identify mutations in type II topoisomerase as a driver of defective proliferation and survival of B2 cells, leading to a B cell immunodeficiency.
Tue Aug 13 00:00:00 UTC 2019
53BP1 cooperation with the REV7-shieldin complex underpins DNA structure-specific NHEJ.
Nature
Ghezraoui H,Oliveira C,Becker JR,Bilham K,Moralli D,Anzilotti C,Fischer R,Deobagkar-Lele M,Sanchiz-Calvo M,Fueyo-Marcos E,Bonham S,Kessler BM,Rottenberg S,Cornall RJ,Green CM,Chapman JR
11-0431 was used in Flow cytometry/Cell sorting to conclude that the 53BP1 pathway therefore comprises distinct double-strand break repair activities within chromatin and single-stranded DNA compartments, which explains both the immunological differences between 53bp1- and Rev7- deficient mice and the context specificity of the pathway.
Wed Aug 01 00:00:00 UTC 2018
53BP1 cooperation with the REV7-shieldin complex underpins DNA structure-specific NHEJ.
Nature
Ghezraoui H,Oliveira C,Becker JR,Bilham K,Moralli D,Anzilotti C,Fischer R,Deobagkar-Lele M,Sanchiz-Calvo M,Fueyo-Marcos E,Bonham S,Kessler BM,Rottenberg S,Cornall RJ,Green CM,Chapman JR
11-0431 was used in Flow cytometry/Cell sorting to conclude that the 53BP1 pathway therefore comprises distinct double-strand break repair activities within chromatin and single-stranded DNA compartments, which explains both the immunological differences between 53bp1- and Rev7- deficient mice and the context specificity of the pathway.
Wed Aug 01 00:00:00 UTC 2018
Inactivation of mTORC1 Signaling in Osterix-Expressing Cells Impairs B-cell Differentiation.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Wang Y,Xiao M,Tao C,Chen J,Wang Z,Yang J,Chen Z,Zou Z,Liu A,Cai D,Jiang Y,Ding C,Li M,Bai X
Published figure using CD43 monoclonal antibody (Product # 11-0431-82) in Flow Cytometry
Sun Apr 01 00:00:00 UTC 2018
ORMDL3 Facilitates the Survival of Splenic B Cells via an ATF6α-Endoplasmic Reticulum Stress-Beclin1 Autophagy Regulatory Pathway.
Journal of immunology (Baltimore, Md. : 1950)
Dang J,Bian X,Ma X,Li J,Long F,Shan S,Yuan Q,Xin Q,Li Y,Gao F,Gong Y,Liu Q
11-0431 was used in Flow cytometry/Cell sorting to study the upregulation of orosomucoid-like 3 in both patients with systemic lupus erythematosus and lupus mice compared with controls.
Fri Sep 01 00:00:00 UTC 2017
Loss of IP3 Receptor-Mediated Ca2+ Release in Mouse B Cells Results in Abnormal B Cell Development and Function.
Journal of immunology (Baltimore, Md. : 1950)
Tang H,Wang H,Lin Q,Fan F,Zhang F,Peng X,Fang X,Liu J,Ouyang K
11-0431 was used in Flow cytometry/Cell sorting to determine the function of 1,4,5-trisphosphate receptor-mediated Ca2+ release in B cells.
Sat Jul 15 00:00:00 UTC 2017
lncRNA requirements for mouse acute myeloid leukemia and normal differentiation.
eLife
Delás MJ,Sabin LR,Dolzhenko E,Knott SR,Munera Maravilla E,Jackson BT,Wild SA,Kovacevic T,Stork EM,Zhou M,Erard N,Lee E,Kelley DR,Roth M,Barbosa IA,Zuber J,Rinn JL,Smith AD,Hannon GJ
11-0431 was used in Flow cytometry/Cell sorting to assess lncRNA function via an in vivo RNAi screen in a model of acute myeloid leukemia.
Wed Sep 06 00:00:00 UTC 2017
Myeloid 12/15-LOX regulates B cell numbers and innate immune antibody levels in vivo.
Wellcome open research
Lauder SN,Tyrrell VJ,Allen-Redpath K,Aldrovandi M,Gray D,Collins P,Jones SA,Taylor PR,O'Donnell V
11-0431 was used in Flow cytometry/Cell sorting to discover that 12/15-LOX mediates alterations in B cell numbers and innate immune antibody generation.
Wed Jan 04 00:00:00 UTC 2017
Alkylator-Induced and Patient-Derived Xenograft Mouse Models of Therapy-Related Myeloid Neoplasms Model Clinical Disease and Suggest the Presence of Multiple Cell Subpopulations with Leukemia Stem Cell Activity.
PloS one
Jonas BA,Johnson C,Gratzinger D,Majeti R
11-0431 was used in Flow cytometry/Cell sorting to report several new t-AML/MDS mouse models that could potentially be used to further define disease pathogenesis and test novel therapeutics.
Wed Jul 26 00:00:00 UTC 2017
Distinctive Mesenchymal-Parenchymal Cell Pairings Govern B Cell Differentiation in the Bone Marrow.
Stem cell reports
Yu VW,Lymperi S,Oki T,Jones A,Swiatek P,Vasic R,Ferraro F,Scadden DT
11-0431 was used in Flow cytometry/Cell sorting to study the notion that bone marrow is a composite of specialised niches, providing distinctive functional units to regulate hematopoiesis.
Tue Aug 09 00:00:00 UTC 2016
The immunosenescence-related gene Zizimin2 is associated with early bone marrow B cell development and marginal zone B cell formation.
Immunity & ageing : I & A
Matsuda T,Yanase S,Takaoka A,Maruyama M
11-0431 was used in Flow cytometry/Cell sorting to show the immunosenescence-related gene Zizimin2 is associated with early bone marrow B cell development and marginal zone B cell formation.
Mon Mar 02 00:00:00 UTC 2015
Rhof promotes murine marginal zone B cell development.
Nagoya journal of medical science
Kishimoto M,Matsuda T,Yanase S,Katsumi A,Suzuki N,Ikejiri M,Takagi A,Ikawa M,Kojima T,Kunishima S,Kiyoi H,Naoe T,Matsushita T,Maruyama M
11-0431 was used in Flow cytometry/Cell sorting to demonstrate that RhoF promotes MZ B cell development in the spleen.
Fri Aug 01 00:00:00 UTC 2014
Rhof promotes murine marginal zone B cell development.
Nagoya journal of medical science
Kishimoto M,Matsuda T,Yanase S,Katsumi A,Suzuki N,Ikejiri M,Takagi A,Ikawa M,Kojima T,Kunishima S,Kiyoi H,Naoe T,Matsushita T,Maruyama M
11-0431 was used in Flow cytometry/Cell sorting to demonstrate that RhoF promotes MZ B cell development in the spleen.
Fri Aug 01 00:00:00 UTC 2014
A mouse model for inducible overexpression of Prdm14 results in rapid-onset and highly penetrant T-cell acute lymphoblastic leukemia (T-ALL).
Disease models & mechanisms
Carofino BL,Ayanga B,Justice MJ
Published figure using CD43 monoclonal antibody (Product # 11-0431-82) in Flow Cytometry
Fri Nov 01 00:00:00 UTC 2013
TDP2-dependent non-homologous end-joining protects against topoisomerase II-induced DNA breaks and genome instability in cells and in vivo.
PLoS genetics
Gómez-Herreros F,Romero-Granados R,Zeng Z,Alvarez-Quilón A,Quintero C,Ju L,Umans L,Vermeire L,Huylebroeck D,Caldecott KW,Cortés-Ledesma F
Published figure using CD43 monoclonal antibody (Product # 11-0431-82) in Flow Cytometry
Thu Jun 13 00:00:00 UTC 2013
Prolactin levels correlate with abnormal B cell maturation in MRL and MRL/lpr mouse models of systemic lupus erythematosus-like disease.
Clinical & developmental immunology
Legorreta-Haquet MV,Flores-Fernández R,Blanco-Favela F,Fuentes-Pananá EM,Chávez-Sánchez L,Hernández-González R,Tesoro-Cruz E,Arriaga-Pizano L,Chávez-Rueda AK
11-0431 was used in Flow cytometry/Cell sorting to determine the level of PRL receptor expression during bone-marrow B-cell development and assess relevance in disease outcomes in lupus-prone murine models.
Mon Aug 04 00:00:00 UTC 2014
IKKα-mediated signaling circuitry regulates early B lymphopoiesis during hematopoiesis.
Blood
Balkhi MY,Willette-Brown J,Zhu F,Chen Z,Liu S,Guttridge DC,Karin M,Hu Y
11-0431-82 was used in Flow Cytometry to demonstrate that IKKα regulates B-lineage commitment via combined canonical and noncanonical NF-κB transcriptional activities to target Pax5 expression during hematopoiesis.
Thu Jun 07 00:00:00 UTC 2012
Conditional TPM3-ALK and NPM-ALK transgenic mice develop reversible ALK-positive early B-cell lymphoma/leukemia.
Blood
Giuriato S,Foisseau M,Dejean E,Felsher DW,Al Saati T,Demur C,Ragab A,Kruczynski A,Schiff C,Delsol G,Meggetto F
11-0431 was used in Flow cytometry/Cell sorting to develop conditional mouse models for anaplastic lymphoma kinase-induced lymphomagenesis.
Thu May 20 00:00:00 UTC 2010
Homologous recombination is necessary for normal lymphocyte development.
Molecular and cellular biology
Caddle LB,Hasham MG,Schott WH,Shirley BJ,Mills KD
11-0431 was used in Flow cytometry/Cell sorting to demonstrate a key role for HR in lymphoid development and suggest that Xrcc2 defects could underlie some human primary immunodeficiencies.
Tue Apr 01 00:00:00 UTC 2008