Product References
A platinum@polymer-catechol nanobraker enables radio-immunotherapy for crippling melanoma tumorigenesis, angiogenesis, and radioresistance.
Bioactive materials
Li W,Yan J,Tian H,Li B,Wang G,Sang W,Zhang Z,Zhang X,Dai Y
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2023
Prolonged dysbiosis and altered immunity under nutritional intervention in a physiological mouse model of severe acute malnutrition.
iScience
Hidalgo-Villeda F,Million M,Defoort C,Vannier T,Svilar L,Lagier M,Wagner C,Arroyo-Portilla C,Chasson L,Luciani C,Bossi V,Gorvel JP,Lelouard H,Tomas J
56-0441-82 was used in Flow cytometry/Cell sorting to provide a preclinical model of SAM and have identified key markers to target with future interventions during the education of the immune system to improve SAM whole defects.
Fri Jun 16 00:00:00 EDT 2023
TRAF6 controls T cell homeostasis by maintaining the equilibrium of MALT1 scaffolding and protease functions.
Frontiers in immunology
O'Neill TJ,Gewies A,Seeholzer T,Krappmann D
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Feb 13 00:00:00 EST 2023
Mitochondrial RNA modifications shape metabolic plasticity in metastasis.
Nature
Delaunay S,Pascual G,Feng B,Klann K,Behm M,Hotz-Wagenblatt A,Richter K,Zaoui K,Herpel E,Münch C,Dietmann S,Hess J,Benitah SA,Frye M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunohistochemistry
Fri Jul 01 00:00:00 EDT 2022
CD153/CD30 signaling promotes age-dependent tertiary lymphoid tissue expansion and kidney injury.
The Journal of clinical investigation
Sato Y,Oguchi A,Fukushima Y,Masuda K,Toriu N,Taniguchi K,Yoshikawa T,Cui X,Kondo M,Hosoi T,Komidori S,Shimizu Y,Fujita H,Jiang L,Kong Y,Yamanashi T,Seita J,Yamamoto T,Toyokuni S,Hamazaki Y,Hattori M,Yoshikai Y,Boor P,Floege J,Kawamoto H,Murakawa Y,Minato N,Yanagita M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Jan 18 00:00:00 EST 2022
FUS-DDIT3 Fusion Oncoprotein Expression Affects JAK-STAT Signaling in Myxoid Liposarcoma.
Frontiers in oncology
Dolatabadi S,Jonasson E,Andersson L,Luna Santamaría M,Lindén M,Österlund T,Åman P,Ståhlberg A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Feb 23 00:00:00 EST 2022
LncRNA ANRIL-mediated miR-181b-5p/S1PR1 axis is involved in the progression of uremic cardiomyopathy through activating T cells.
Scientific reports
Xu Y,Cao L,Ji S,Shen W
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Oct 27 00:00:00 EDT 2022
Isolation and characterization mesenchymal stem cells from red panda (Ailurus fulgens styani) endometrium.
Conservation physiology
Wang DH,Wu XM,Chen JS,Cai ZG,An JH,Zhang MY,Li Y,Li FP,Hou R,Liu YL
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sat Jan 01 00:00:00 EST 2022
WDR82-binding long noncoding RNA lncEry controls mouse erythroid differentiation and maturation.
The Journal of experimental medicine
Yang S,Sun G,Wu P,Chen C,Kuang Y,Liu L,Zheng Z,He Y,Gu Q,Lu T,Zhu C,Wang F,Gou F,Yang Z,Zhao X,Yuan S,Yang L,Lu S,Li Y,Lv X,Dong F,Ma Y,Yu J,Ng LG,Shi L,Liu J,Shi L,Cheng T,Cheng H
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Apr 04 00:00:00 EDT 2022
Multivalent state transitions shape the intratumoral composition of small cell lung carcinoma.
Science advances
Gopal P,Petty A,Rogacki K,Bera T,Bareja R,Peacock CD,Abazeed ME
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Dec 14 00:00:00 EST 2022
High-Contrast Stimulation Potentiates the Neurotrophic Properties of Müller Cells and Suppresses Their Pro-Inflammatory Phenotype.
International journal of molecular sciences
Zloh M,Kutilek P,Stofkova A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunohistochemistry
Wed Aug 03 00:00:00 EDT 2022
CXCR4, CXCR5 and CD44 May Be Involved in Homing of Lymphoma Cells into the Eye in a Patient Derived Xenograft Homing Mouse Model for Primary Vitreoretinal Lymphoma.
International journal of molecular sciences
Babst N,Isbell LK,Rommel F,Tura A,Ranjbar M,Grisanti S,Tschuch C,Schueler J,Doostkam S,Reinacher PC,Duyster J,Kakkassery V,von Bubnoff N
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunohistochemistry
Tue Oct 04 00:00:00 EDT 2022
Lin28B-high breast cancer cells promote immune suppression in the lung pre-metastatic niche via exosomes and support cancer progression.
Nature communications
Qi M,Xia Y,Wu Y,Zhang Z,Wang X,Lu L,Dai C,Song Y,Xu K,Ji W,Zhan L
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Feb 16 00:00:00 EST 2022
RNF4~RGMb~BMP6 axis required for osteogenic differentiation and cancer cell survival.
Cell death & disease
Novak R,Ahmad YA,Timaner M,Bitman-Lotan E,Oknin-Vaisman A,Horwitz R,Hartmann O,Reissland M,Buck V,Rosenfeldt M,Nikomarov D,Diefenbacher ME,Shaked Y,Orian A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sat Sep 24 00:00:00 EDT 2022
CD44 Promotes Breast Cancer Metastasis through AKT-Mediated Downregulation of Nuclear FOXA2.
Biomedicines
Vadhan A,Hou MF,Vijayaraghavan P,Wu YC,Hu SC,Wang YM,Cheng TL,Wang YY,Yuan SF
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Oct 05 00:00:00 EDT 2022
Circ_RNF13 Regulates the Stemness and Chemosensitivity of Colorectal Cancer by Transcriptional Regulation of DDX27 Mediated by TRIM24 Stabilization.
Cancers
Guo Y,Hu G,Tian B,Ma M,Long F,Chen M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Dec 16 00:00:00 EST 2022
Smad4 and p53 synergize in suppressing autochthonous intestinal cancer.
Cancer medicine
Park JW,Seo MJ,Cho KS,Kook MC,Jeong JM,Roh SG,Cho SY,Cheon JH,Kim HK
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sun May 01 00:00:00 EDT 2022
Amphiphile-CpG vaccination induces potent lymph node activation and COVID-19 immunity in mice and non-human primates.
NPJ vaccines
Seenappa LM,Jakubowski A,Steinbuck MP,Palmer E,Haqq CM,Carter C,Fontenot J,Villinger F,McNeil LK,DeMuth PC
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Oct 28 00:00:00 EDT 2022
Hobit and Blimp-1 instruct the differentiation of iNKT cells into resident-phenotype lymphocytes after lineage commitment.
European journal of immunology
Kragten NA,Taggenbrock RL,Parga Vidal L,van Lier RA,Stark R,van Gisbergen KP
56-0441-82 was used in Flow cytometry/Cell sorting to conclude that memory differentiation and the establishment of residency occur after lineage commitment through a Hobit and Blimp-1-driven transcriptional program.
Tue Mar 01 00:00:00 EST 2022
Adipocyte CD1d Gene Transfer Induces T Cell Expansion and Adipocyte Inflammation in CD1d Knockout Mice.
Journal of immunology (Baltimore, Md. : 1950)
Xiao R,Mansour AG,Huang W,Hassan QN,Wilkins RK,Komatineni SV,Bates R,Ali S,Chrislip LA,Queen NJ,Ma S,Yu J,Lordo MR,Mundy-Bosse BL,Caligiuri MA,Cao L
56-0441-82 was used in Flow cytometry/Cell sorting to report that adeno-associated virus-mediated gene transfer of CD1d to adipocytes in CD1d KO mice fails to rescue iNKT cells but leads to massive and selective expansion of T cells within adipose tissue, particularly CD8+ T effector cells, that is associated with adipocyte NLRP3 inflammasome activation, dysregulation of adipocyte functional genes, and upregulation of apoptotic pathway proteins.
Sun May 01 00:00:00 EDT 2022
Age-Associated Changes to Lymph Node Fibroblastic Reticular Cells.
Frontiers in aging
Kwok T,Medovich SC,Silva-Junior IA,Brown EM,Haug JC,Barrios MR,Morris KA,Lancaster JN
56-0441-82 was used in Flow cytometry/Cell sorting to address the controversy as to whether aging impacts the composition lymph node stroma and supports a model in which impaired differentiation of lymph node fibroblasts and increased fibrosis inhibits the interactions necessary for naive T cell homeostasis.
Sat Jul 16 00:00:00 EDT 2022
Murine model of colonization with fungal pathogen Candida auris to explore skin tropism, host risk factors and therapeutic strategies.
Cell host & microbe
Huang X,Hurabielle C,Drummond RA,Bouladoux N,Desai JV,Sim CK,Belkaid Y,Lionakis MS,Segre JA
56-0441-82 was used in Flow Cytometry to develop a murine skin topical exposure model for C. auris to dissect risk factors for colonization and to test interventions that might protect patients.
Wed Feb 10 00:00:00 EST 2021
NF-κB-inducing kinase maintains T cell metabolic fitness in antitumor immunity.
Nature immunology
Gu M,Zhou X,Sohn JH,Zhu L,Jie Z,Yang JY,Zheng X,Xie X,Yang J,Shi Y,Brightbill HD,Kim JB,Wang J,Cheng X,Sun SC
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Disruption of the MSL complex inhibits tumour maintenance by exacerbating chromosomal instability.
Nature cell biology
Monserrat J,Morales Torres C,Richardson L,Wilson TS,Patel H,Domart MC,Horswell S,Song OR,Jiang M,Crawford M,Bui M,Dalal Y,Scaffidi P
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
A TLR7 antagonist restricts interferon-dependent and -independent immunopathology in a mouse model of severe influenza.
The Journal of experimental medicine
Rappe JCF,Finsterbusch K,Crotta S,Mack M,Priestnall SL,Wack A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Nov 01 00:00:00 EDT 2021
Chronic T cell proliferation in brains after stroke could interfere with the efficacy of immunotherapies.
The Journal of experimental medicine
Heindl S,Ricci A,Carofiglio O,Zhou Q,Arzberger T,Lenart N,Franzmeier N,Hortobagyi T,Nelson PT,Stowe AM,Denes A,Edbauer D,Liesz A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Aug 02 00:00:00 EDT 2021
Stage-specific action of Runx1 and GATA3 controls silencing of PU.1 expression in mouse pro-T cells.
The Journal of experimental medicine
Hosokawa H,Koizumi M,Masuhara K,Romero-Wolf M,Tanaka T,Nakayama T,Rothenberg EV
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Aug 02 00:00:00 EDT 2021
Low immunogenicity of malaria pre-erythrocytic stages can be overcome by vaccination.
EMBO molecular medicine
Müller K,Gibbins MP,Roberts M,Reyes-Sandoval A,Hill AVS,Draper SJ,Matuschewski K,Silvie O,Hafalla JCR
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Apr 09 00:00:00 EDT 2021
Senescent T Cell Induces Brown Adipose Tissue "Whitening" Via Secreting IFN-γ.
Frontiers in cell and developmental biology
Pan XX,Yao KL,Yang YF,Ge Q,Zhang R,Gao PJ,Ruan CC,Wu F
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Mar 23 00:00:00 EDT 2021
Trichinella spiralis Paramyosin Induces Colonic Regulatory T Cells to Mitigate Inflammatory Bowel Disease.
Frontiers in cell and developmental biology
Hao C,Wang W,Zhan B,Wang Z,Huang J,Sun X,Zhu X
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Aug 03 00:00:00 EDT 2021
Downregulation of SLC27A6 by DNA Hypermethylation Promotes Proliferation but Suppresses Metastasis of Nasopharyngeal Carcinoma Through Modulating Lipid Metabolism.
Frontiers in oncology
Zhong X,Yang Y,Li B,Liang P,Huang Y,Zheng Q,Wang Y,Xiao X,Mo Y,Zhang Z,Zhou X,Huang G,Zhao W
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Jan 21 00:00:00 EST 2022
Thrombospondin-2 spatiotemporal expression in skeletal fractures.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Zondervan RL,Jenkins DC,Reicha JD,Hankenson KD
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
The white matter is a pro-differentiative niche for glioblastoma.
Nature communications
Brooks LJ,Clements MP,Burden JJ,Kocher D,Richards L,Devesa SC,Zakka L,Woodberry M,Ellis M,Jaunmuktane Z,Brandner S,Morrison G,Pollard SM,Dirks PB,Marguerat S,Parrinello S
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunohistochemistry
Mon Apr 12 00:00:00 EDT 2021
Differential activation of Ca2+ influx channels modulate stem cell potency, their proliferation/viability and tissue regeneration.
NPJ Regenerative medicine
Ahamad N,Sun Y,Nascimento Da Conceicao V,Xavier Paul Ezhilan CRD,Natarajan M,Singh BB
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunohistochemistry
Wed Oct 20 00:00:00 EDT 2021
Neuroimmune Consequences of eIF4E Phosphorylation on Chemotherapy-Induced Peripheral Neuropathy.
Frontiers in immunology
Agalave NM,Mody PH,Szabo-Pardi TA,Jeong HS,Burton MD
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Sep 20 00:00:00 EDT 2021
Prophylactic efficacy against Mycobacterium tuberculosis using ID93 and lipid-based adjuvant formulations in the mouse model.
PloS one
Baldwin SL,Reese VA,Larsen SE,Beebe E,Guderian J,Orr MT,Fox CB,Reed SG,Coler RN
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Oct 13 00:00:00 EDT 2021
In Vitro Anti-cancer Activity of Adipose-Derived Mesenchymal Stem Cells Increased after Infection with Oncolytic Reovirus.
Advanced pharmaceutical bulletin
Babaei A,Bannazadeh Baghi H,Nezhadi A,Jamalpoor Z
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Valproic acid stimulates myogenesis in pluripotent stem cell-derived mesodermal progenitors in a NOTCH-dependent manner.
Cell death & disease
Breuls N,Giarratana N,Yedigaryan L,Garrido GM,Carai P,Heymans S,Ranga A,Deroose C,Sampaolesi M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Jul 05 00:00:00 EDT 2021
miR-34a mimic or pre-mir-34a, which is the better option for cancer therapy? KatoIII as a model to study miRNA action in human gastric cancer cells.
Cancer cell international
Jafari N,Abediankenari S,Hossein-Nataj H
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Mar 19 00:00:00 EDT 2021
Immuno-Electron and Confocal Laser Scanning Microscopy of the Glycocalyx.
Biology
Twamley SG,Stach A,Heilmann H,Söhl-Kielczynski B,Stangl V,Ludwig A,Münster-Wandowski A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunocytochemistry
Tue May 04 00:00:00 EDT 2021
Immuno-Electron and Confocal Laser Scanning Microscopy of the Glycocalyx.
Biology
Twamley SG,Stach A,Heilmann H,Söhl-Kielczynski B,Stangl V,Ludwig A,Münster-Wandowski A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunocytochemistry
Tue May 04 00:00:00 EDT 2021
PI3Kδ coordinates transcriptional, chromatin, and metabolic changes to promote effector CD8+ T cells at the expense of central memory.
Cell reports
Cannons JL,Villarino AV,Kapnick SM,Preite S,Shih HY,Gomez-Rodriguez J,Kaul Z,Shibata H,Reilley JM,Huang B,Handon R,McBain IT,Gossa S,Wu T,Su HC,McGavern DB,O'Shea JJ,McGuire PJ,Uzel G,Schwartzberg PL
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Oct 12 00:00:00 EDT 2021
MiR-125b-5p Is Involved in Sorafenib Resistance through Ataxin-1-Mediated Epithelial-Mesenchymal Transition in Hepatocellular Carcinoma.
Cancers
Hirao A,Sato Y,Tanaka H,Nishida K,Tomonari T,Hirata M,Bando M,Kida Y,Tanaka T,Kawaguchi T,Wada H,Taniguchi T,Okamoto K,Miyamoto H,Muguruma N,Tanahashi T,Takayama T
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Sep 30 00:00:00 EDT 2021
Synchronous effects of targeted mitochondrial complex I inhibitors on tumor and immune cells abrogate melanoma progression.
iScience
AbuEid M,McAllister DM,McOlash L,Harwig MC,Cheng G,Drouillard D,Boyle KA,Hardy M,Zielonka J,Johnson BD,Hill RB,Kalyanaraman B,Dwinell MB
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Jun 25 00:00:00 EDT 2021
Theranostic near-infrared-IIb emitting nanoprobes for promoting immunogenic radiotherapy and abscopal effects against cancer metastasis.
Nature communications
Li H,Wang M,Huang B,Zhu SW,Zhou JJ,Chen DR,Cui R,Zhang M,Sun ZJ
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Dec 09 00:00:00 EST 2021
Therapeutic Potential of Mesenchymal Stem Cells in a Pre-Clinical Model of Diabetic Kidney Disease and Obesity.
International journal of molecular sciences
Sávio-Silva C,Soinski-Sousa PE,Simplício-Filho A,Bastos RMC,Beyerstedt S,Rangel ÉB
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Feb 04 00:00:00 EST 2021
Attenuation of apoptotic cell detection triggers thymic regeneration after damage.
Cell reports
Kinsella S,Evandy CA,Cooper K,Iovino L,deRoos PC,Hopwo KS,Granadier DW,Smith CW,Rafii S,Dudakov JA
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Oct 05 00:00:00 EDT 2021
Protocols for isolation and characterization of mouse placental hemogenic endothelial cells.
STAR protocols
Liang G,Huang B,Wang F,Liu F
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Dec 17 00:00:00 EST 2021
Near-infrared photoimmunotherapy targeting human-EGFR in a mouse tumor model simulating current and future clinical trials.
EBioMedicine
Okada R,Furusawa A,Vermeer DW,Inagaki F,Wakiyama H,Kato T,Nagaya T,Choyke PL,Spanos WC,Allen CT,Kobayashi H
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sat May 01 00:00:00 EDT 2021
c-FOS drives reversible basal to squamous cell carcinoma transition.
Cell reports
Kuonen F,Li NY,Haensel D,Patel T,Gaddam S,Yerly L,Rieger K,Aasi S,Oro AE
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunocytochemistry
Tue Oct 05 00:00:00 EDT 2021
CD95/Fas protects triple negative breast cancer from anti-tumor activity of NK cells.
iScience
Qadir AS,Guégan JP,Ginestier C,Chaibi A,Bessede A,Charafe-Jauffret E,Macario M,Lavoué V,Rouge TM,Law C,Vilker J,Wang H,Stroup E,Schipma MJ,Bridgeman B,Murmann AE,Ji Z,Legembre P,Peter ME
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunocytochemistry
Fri Nov 19 00:00:00 EST 2021
3-hydroxyanthranic acid increases the sensitivity of hepatocellular carcinoma to sorafenib by decreasing tumor cell stemness.
Cell death discovery
Gan G,Shi Z,Liu D,Zhang S,Zhu H,Wang Y,Mi J
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Jul 06 00:00:00 EDT 2021
Serglycin induces osteoclastogenesis and promotes tumor growth in giant cell tumor of bone.
Cell death & disease
He Y,Cheng D,Lian C,Liu Y,Luo W,Wang Y,Ma C,Wu Q,Tian P,He D,Jia Z,Lv X,Zhang X,Pan Z,Lu J,Xiao Y,Zhang P,Liang Y,Yang Q,Hu G
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Neutralization
Thu Sep 23 00:00:00 EDT 2021
A CD10-OGP Membrane Peptolytic Signaling Axis in Fibroblasts Regulates Lipid Metabolism of Cancer Stem Cells via SCD1.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Yu S,Lu Y,Su A,Chen J,Li J,Zhou B,Liu X,Xia Q,Li Y,Li J,Huang M,Ye Y,Zhao Q,Jiang S,Yan X,Wang X,Di C,Pan J,Su S
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2021
Type 2 diabetic mice enter a state of spontaneous hibernation-like suspended animation following accumulation of uric acid.
The Journal of biological chemistry
Zhao Y,Cheng R,Zhao Y,Ge W,Yang Y,Ding Z,Xu X,Wang Z,Wu Z,Zhang J
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2021
A nanounit strategy reverses immune suppression of exosomal PD-L1 and is associated with enhanced ferroptosis.
Nature communications
Wang G,Xie L,Li B,Sang W,Yan J,Li J,Tian H,Li W,Zhang Z,Tian Y,Dai Y
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Sep 30 00:00:00 EDT 2021
GPR120 induces regulatory dendritic cells by inhibiting HK2-dependent glycolysis to alleviate fulminant hepatic failure.
Cell death & disease
Yu H,Yang W,Huang J,Miao X,Wang B,Ren X,Gu Y,Wang Q,Ding X,Guo X,Qian F,Zhang Y,Xu H,Zheng L,Jin M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Dec 16 00:00:00 EST 2021
Graded RhoA GTPase Expression in Treg Cells Distinguishes Tumor Immunity From Autoimmunity.
Frontiers in immunology
Kalim KW,Yang JQ,Modur V,Nguyen P,Li Y,Zheng Y,Guo F
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Jan 11 00:00:00 EST 2022
Analysis of T cells in mouse lymphoid tissue and blood with flow cytometry.
STAR protocols
Skordos I,Demeyer A,Beyaert R
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Mar 19 00:00:00 EDT 2021
IL4I1 Is a Metabolic Immune Checkpoint that Activates the AHR and Promotes Tumor Progression.
Cell
Sadik A,Somarribas Patterson LF,Öztürk S,Mohapatra SR,Panitz V,Secker PF,Pfänder P,Loth S,Salem H,Prentzell MT,Berdel B,Iskar M,Faessler E,Reuter F,Kirst I,Kalter V,Foerster KI,Jäger E,Guevara CR,Sobeh M,Hielscher T,Poschet G,Reinhardt A,Hassel JC,Zapatka M,Hahn U,von Deimling A,Hopf C,Schlichting R,Escher BI,Burhenne J,Haefeli WE,Ishaque N,Böhme A,Schäuble S,Thedieck K,Trump S,Seiffert M,Opitz CA
56-0441 was used in Flow cytometry/Cell sorting to show that IL4I1 blockade opens new avenues for cancer therapy.
Thu Sep 03 00:00:00 EDT 2020
Antagonistic Inflammatory Phenotypes Dictate Tumor Fate and Response to Immune Checkpoint Blockade.
Immunity
Bonavita E,Bromley CP,Jonsson G,Pelly VS,Sahoo S,Walwyn-Brown K,Mensurado S,Moeini A,Flanagan E,Bell CR,Chiang SC,Chikkanna-Gowda CP,Rogers N,Silva-Santos B,Jaillon S,Mantovani A,Reis e Sousa C,Guerra N,Davis DM,Zelenay S
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Dec 15 00:00:00 EST 2020
An Integrated Epigenomic and Transcriptomic Map of Mouse and Human αβ T Cell Development.
Immunity
Chopp LB,Gopalan V,Ciucci T,Ruchinskas A,Rae Z,Lagarde M,Gao Y,Li C,Bosticardo M,Pala F,Livak F,Kelly MC,Hannenhalli S,Bosselut R
56-0441-82 was used in Flow Cytometry to identify mouse and human αβ thymocyte developmental trajectories.
Tue Dec 15 00:00:00 EST 2020
An Integrated Epigenomic and Transcriptomic Map of Mouse and Human αβ T Cell Development.
Immunity
Chopp LB,Gopalan V,Ciucci T,Ruchinskas A,Rae Z,Lagarde M,Gao Y,Li C,Bosticardo M,Pala F,Livak F,Kelly MC,Hannenhalli S,Bosselut R
56-0441-82 was used in Flow Cytometry to identify mouse and human αβ thymocyte developmental trajectories.
Tue Dec 15 00:00:00 EST 2020
Pharmacological Activation of Pyruvate Kinase M2 Inhibits CD4+ T Cell Pathogenicity and Suppresses Autoimmunity.
Cell metabolism
Angiari S,Runtsch MC,Sutton CE,Palsson-McDermott EM,Kelly B,Rana N,Kane H,Papadopoulou G,Pearce EL,Mills KHG,O'Neill LAJ
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Feb 04 00:00:00 EST 2020
Transfer of metastatic traits via miR-200c in extracellular vesicles derived from colorectal cancer stem cells is inhibited by atractylenolide I.
Clinical and translational medicine
Tang D,Xu X,Ying J,Xie T,Cao G
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2020
Cancer cells educate natural killer cells to a metastasis-promoting cell state.
The Journal of cell biology
Chan IS,Knútsdóttir H,Ramakrishnan G,Padmanaban V,Warrier M,Ramirez JC,Dunworth M,Zhang H,Jaffee EM,Bader JS,Ewald AJ
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunocytochemistry
Mon Sep 07 00:00:00 EDT 2020
Distinct roles of PIK3CA in the enrichment and maintenance of cancer stem cells in head and neck squamous cell carcinoma.
Molecular oncology
Chen X,Cao Y,Sedhom W,Lu L,Liu Y,Wang H,Oka M,Bornstein S,Said S,Song J,Lu SL
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2020
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
Dexamethasone inhibits stemness maintenance and enhances chemosensitivity of hepatocellular carcinoma stem cells by inducing deSUMOylation of HIF‑1α and Oct4.
International journal of oncology
Jiang Z,Zhang C,Liu X,Ma X,Bian X,Xiao X,Gao R,Sun Y,Wu W,Zhao P
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2020
Extracellular HMGB-1 activates inflammatory signaling in tendon cells and tissues.
Therapeutic advances in chronic disease
Zhang C,Gu X,Zhao G,Wang W,Shao J,Zhu J,Yuan T,Sun J,Nie D,Zhou Y
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sun Apr 17 00:00:00 EDT 2022
Explant culture: A relevant tool for the study of telocytes.
Cell biology international
Sanches BDA,Maldarine JDS,Tamarindo GH,Da Silva ADT,Lima MLD,Rahal P,Góes RM,Taboga SR,Carvalho HF
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunocytochemistry
Tue Dec 01 00:00:00 EST 2020
Influenza NG-34 T cell conserved epitope adjuvanted with CAF01 as a possible influenza vaccine candidate.
Veterinary research
Sisteré-Oró M,Pedersen GK,Córdoba L,López-Serrano S,Christensen D,Darji A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Apr 20 00:00:00 EDT 2020
miR‑210 enhances mesenchymal stem cell‑modulated neural precursor cell migration.
Molecular medicine reports
Wang F,Zhu J,Zheng J,Duan W,Zhou Z
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2020
Identification, characterization and microRNA expression profiling of side population cells in human oral squamous cell carcinoma Tca8113 cell lines.
Molecular medicine reports
Luo W,Liu RS,E LL,Bai Y,Kong XP,Liu HW,Wu H,Liu HC
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2020
Resveratrol rescues TNF‑α‑induced inhibition of osteogenesis in human periodontal ligament stem cells via the ERK1/2 pathway.
Molecular medicine reports
Yuan J,Wang X,Ma D,Gao H,Zheng D,Zhang J
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri May 01 00:00:00 EDT 2020
Chronic expression of p16INK4a in the epidermis induces Wnt-mediated hyperplasia and promotes tumor initiation.
Nature communications
Azazmeh N,Assouline B,Winter E,Ruppo S,Nevo Y,Maly A,Meir K,Witkiewicz AK,Cohen J,Rizou SV,Pikarsky E,Luxenburg C,Gorgoulis VG,Ben-Porath I
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunohistochemistry
Mon Jun 01 00:00:00 EDT 2020
Critical role of WNK1 in MYC-dependent early mouse thymocyte development.
eLife
Köchl R,Vanes L,Llorian Sopena M,Chakravarty P,Hartweger H,Fountain K,White A,Cowan J,Anderson G,Tybulewicz VL
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Oct 14 00:00:00 EDT 2020
Inhibition of IRF4 in dendritic cells by PRR-independent and -dependent signals inhibit Th2 and promote Th17 responses.
eLife
Lee J,Zhang J,Chung YJ,Kim JH,Kook CM,González-Navajas JM,Herdman DS,Nürnberg B,Insel PA,Corr M,Mo JH,Tao A,Yasuda K,Rifkin IR,Broide DH,Sciammas R,Webster NJ,Raz E
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Feb 04 00:00:00 EST 2020
Effect of Combining Low Temperature Plasma, Negative Pressure Wound Therapy, and Bone Marrow Mesenchymal Stem Cells on an Acute Skin Wound Healing Mouse Model.
International journal of molecular sciences
Cui HS,Joo SY,Cho YS,Park JH,Kim JB,Seo CH
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sat May 23 00:00:00 EDT 2020
Targeted expression profiling reveals distinct stages of early canine fibroblast reprogramming are regulated by 2-oxoglutarate hydroxylases.
Stem cell research & therapy
Tobias IC,Kao MC,Parmentier T,Hunter H,LaMarre J,Betts DH
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunocytochemistry
Wed Dec 09 00:00:00 EST 2020
Targeted expression profiling reveals distinct stages of early canine fibroblast reprogramming are regulated by 2-oxoglutarate hydroxylases.
Stem cell research & therapy
Tobias IC,Kao MC,Parmentier T,Hunter H,LaMarre J,Betts DH
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunocytochemistry
Wed Dec 09 00:00:00 EST 2020
Chemotherapeutic Stress Influences Epithelial-Mesenchymal Transition and Stemness in Cancer Stem Cells of Triple-Negative Breast Cancer.
International journal of molecular sciences
Li X,Strietz J,Bleilevens A,Stickeler E,Maurer J
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Jan 08 00:00:00 EST 2020
Inhibition of EZH2 ameliorates bacteria-induced liver injury by repressing RUNX1 in dendritic cells.
Cell death & disease
Wang Y,Wang Q,Wang B,Gu Y,Yu H,Yang W,Ren X,Qian F,Zhao X,Xiao Y,Zhang Y,Jin M,Zhu M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2020
Diacerein treatment prevents colitis-associated cancer in mice.
World journal of clinical oncology
Paulino DSM,Mendes MCS,Camargo JA,Brambilla SR,Wood Dos Santos T,Ribeiro ML,Carvalheira JBC
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunohistochemistry
Thu Sep 24 00:00:00 EDT 2020
Neurokinin-1 Receptor Signaling Is Required for Efficient Ca2+ Flux in T-Cell-Receptor-Activated T Cells.
Cell reports
Morelli AE,Sumpter TL,Rojas-Canales DM,Bandyopadhyay M,Chen Z,Tkacheva O,Shufesky WJ,Wallace CT,Watkins SC,Berger A,Paige CJ,Falo LD,Larregina AT
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Mar 10 00:00:00 EDT 2020
Graphene-based 2D constructs for enhanced fibroblast support.
PloS one
Safina I,Bourdo SE,Algazali KM,Kannarpady G,Watanabe F,Vang KB,Biris AS
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunocytochemistry
Tue Aug 04 00:00:00 EDT 2020
Tumor Cell Associated Hyaluronan-CD44 Signaling Promotes Pro-Tumor Inflammation in Breast Cancer.
Cancers
Witschen PM,Chaffee TS,Brady NJ,Huggins DN,Knutson TP,LaRue RS,Munro SA,Tiegs L,McCarthy JB,Nelson AC,Schwertfeger KL
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri May 22 00:00:00 EDT 2020
Mucosal-associated invariant T cells promote inflammation and intestinal dysbiosis leading to metabolic dysfunction during obesity.
Nature communications
Toubal A,Kiaf B,Beaudoin L,Cagninacci L,Rhimi M,Fruchet B,da Silva J,Corbett AJ,Simoni Y,Lantz O,Rossjohn J,McCluskey J,Lesnik P,Maguin E,Lehuen A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Jul 24 00:00:00 EDT 2020
Mucosal-associated invariant T cells promote inflammation and intestinal dysbiosis leading to metabolic dysfunction during obesity.
Nature communications
Toubal A,Kiaf B,Beaudoin L,Cagninacci L,Rhimi M,Fruchet B,da Silva J,Corbett AJ,Simoni Y,Lantz O,Rossjohn J,McCluskey J,Lesnik P,Maguin E,Lehuen A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Jul 24 00:00:00 EDT 2020
Utility Evaluation of Porcine Enteroids as PDCoV Infection Model in vitro.
Frontiers in microbiology
Luo H,Zheng J,Chen Y,Wang T,Zhang Z,Shan Y,Xu J,Yue M,Fang W,Li X
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
Head and Neck Cancer Stem Cell-Enriched Spheroid Model for Anticancer Compound Screening.
Cells
Goričan L,Gole B,Potočnik U
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Jul 16 00:00:00 EDT 2020
Head and Neck Cancer Stem Cell-Enriched Spheroid Model for Anticancer Compound Screening.
Cells
Goričan L,Gole B,Potočnik U
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Jul 16 00:00:00 EDT 2020
Obesity-Altered Adipose Stem Cells Promote Radiation Resistance of Estrogen Receptor Positive Breast Cancer through Paracrine Signaling.
International journal of molecular sciences
Sabol RA,Villela VA,Denys A,Freeman BT,Hartono AB,Wise RM,Harrison MAA,Sandler MB,Hossain F,Miele L,Bunnell BA
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Apr 15 00:00:00 EDT 2020
Cancer-Specific Loss of p53 Leads to a Modulation of Myeloid and T Cell Responses.
Cell reports
Blagih J,Zani F,Chakravarty P,Hennequart M,Pilley S,Hobor S,Hock AK,Walton JB,Morton JP,Gronroos E,Mason S,Yang M,McNeish I,Swanton C,Blyth K,Vousden KH
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Jan 14 00:00:00 EST 2020
Shp1 Loss Enhances Macrophage Effector Function and Promotes Anti-Tumor Immunity.
Frontiers in immunology
Myers DR,Abram CL,Wildes D,Belwafa A,Welsh AMN,Schulze CJ,Choy TJ,Nguyen T,Omaque N,Hu Y,Singh M,Hansen R,Goldsmith MA,Quintana E,Smith JAM,Lowell CA
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Jun 14 00:00:00 EDT 2021
CCL5 derived from tumor-associated macrophages promotes prostate cancer stem cells and metastasis via activating β-catenin/STAT3 signaling.
Cell death & disease
Huang R,Wang S,Wang N,Zheng Y,Zhou J,Yang B,Wang X,Zhang J,Guo L,Wang S,Chen Z,Wang Z,Xiang S
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Apr 16 00:00:00 EDT 2020
Profiling and Targeting of Energy and Redox Metabolism in Grade 2 Bladder Cancer Cells with Different Invasiveness Properties.
Cells
Pasquale V,Ducci G,Campioni G,Ventrici A,Assalini C,Busti S,Vanoni M,Vago R,Sacco E
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Dec 11 00:00:00 EST 2020
Phosphatase SHP1 impedes mesenchymal stromal cell immunosuppressive capacity modulated by JAK1/STAT3 and P38 signals.
Cell & bioscience
Jiang M,Ye J,Wang X,Li N,Wang Y,Shi Y
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Apr 14 00:00:00 EDT 2022
Requirements for the differentiation of innate T-bethigh memory-phenotype CD4+ T lymphocytes under steady state.
Nature communications
Kawabe T,Yi J,Kawajiri A,Hilligan K,Fang D,Ishii N,Yamane H,Zhu J,Jankovic D,Kim KS,Trinchieri G,Sher A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Jul 06 00:00:00 EDT 2020
Localized cocktail chemoimmunotherapy after in situ gelation to trigger robust systemic antitumor immune responses.
Science advances
Chao Y,Liang C,Tao H,Du Y,Wu D,Dong Z,Jin Q,Chen G,Xu J,Xiao Z,Chen Q,Wang C,Chen J,Liu Z
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
The Bone Marrow Protects and Optimizes Immunological Memory during Dietary Restriction.
Cell
Collins N,Han SJ,Enamorado M,Link VM,Huang B,Moseman EA,Kishton RJ,Shannon JP,Dixit D,Schwab SR,Hickman HD,Restifo NP,McGavern DB,Schwartzberg PL,Belkaid Y
56-0441-82 was used in Flow Cytometry to show that memory T cells collapsed in secondary lymphoid organs in the context of dietary restriction (DR) but dramatically accumulated within the bone marrow (BM), where they adopted a state associated with energy conservation.
Thu Aug 22 00:00:00 EDT 2019
A Thpok-Directed Transcriptional Circuitry Promotes Bcl6 and Maf Expression to Orchestrate T Follicular Helper Differentiation.
Immunity
Vacchio MS,Ciucci T,Gao Y,Watanabe M,Balmaceno-Criss M,McGinty MT,Huang A,Xiao Q,McConkey C,Zhao Y,Shetty J,Tran B,Pepper M,Vahedi G,Jenkins MK,McGavern DB,Bosselut R
56-0441-82 was used in Flow Cytometry to identify a transcriptional program that links the CD4+ lineage with Tfh differentiation, a limiting factor for efficient B cell responses.
Tue Sep 17 00:00:00 EDT 2019
Toxin-Triggered Interleukin-1 Receptor Signaling Enables Early-Life Discrimination of Pathogenic versus Commensal Skin Bacteria.
Cell host & microbe
Leech JM,Dhariwala MO,Lowe MM,Chu K,Merana GR,Cornuot C,Weckel A,Ma JM,Leitner EG,Gonzalez JR,Vasquez KS,Diep BA,Scharschmidt TC
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Dec 11 00:00:00 EST 2019
A20 in Myeloid Cells Protects Against Hypertension by Inhibiting Dendritic Cell-Mediated T-Cell Activation.
Circulation research
Lu X,Rudemiller NP,Wen Y,Ren J,Hammer GE,Griffiths R,Privratsky JR,Yang B,Sparks MA,Crowley SD
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Dec 06 00:00:00 EST 2019
A Novel Form of 4-1BBL Prevents Cancer Development via Nonspecific Activation of CD4+ T and Natural Killer Cells.
Cancer research
Barsoumian HB,Batra L,Shrestha P,Bowen WS,Zhao H,Egilmez NK,Gomez-Gutierrez JG,Yolcu ES,Shirwan H
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Feb 15 00:00:00 EST 2019
Clonal copy-number mosaicism in autoreactive T lymphocytes in diabetic NOD mice.
Genome research
Alriyami M,Marchand L,Li Q,Du X,Olivier M,Polychronakos C
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sun Dec 01 00:00:00 EST 2019
The ROP16III-dependent early immune response determines the subacute CNS immune response and type III Toxoplasma gondii survival.
PLoS pathogens
Tuladhar S,Kochanowsky JA,Bhaskara A,Ghotmi Y,Chandrasekaran S,Koshy AA
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2019
In vitro Characteristics of Heterogeneous Equine Hoof Progenitor Cell Isolates.
Frontiers in bioengineering and biotechnology
Yang Q,Pinto VMR,Duan W,Paxton EE,Dessauer JH,Ryan W,Lopez MJ
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
In vitro Characteristics of Heterogeneous Equine Hoof Progenitor Cell Isolates.
Frontiers in bioengineering and biotechnology
Yang Q,Pinto VMR,Duan W,Paxton EE,Dessauer JH,Ryan W,Lopez MJ
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
Leptin Induces Proadipogenic and Proinflammatory Signaling in Adipocytes.
Frontiers in endocrinology
Palhinha L,Liechocki S,Hottz ED,Pereira JADS,de Almeida CJ,Moraes-Vieira PMM,Bozza PT,Maya-Monteiro CM
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Jan 13 00:00:00 EST 2020
Anisomycin inhibits angiogenesis in ovarian cancer by attenuating the molecular sponge effect of the lncRNA‑Meg3/miR‑421/PDGFRA axis.
International journal of oncology
Ye W,Ni Z,Yicheng S,Pan H,Huang Y,Xiong Y,Liu T
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sun Dec 01 00:00:00 EST 2019
TCR Affinity Biases Th Cell Differentiation by Regulating CD25, Eef1e1, and Gbp2.
Journal of immunology (Baltimore, Md. : 1950)
Kotov DI,Mitchell JS,Pengo T,Ruedl C,Way SS,Langlois RA,Fife BT,Jenkins MK
56-0441-82 was used in Flow Cytometry to study TCR affinity and influence on regulation of genes in naive T cells that control the differentiation process in cells.
Wed May 01 00:00:00 EDT 2019
Hypoxia changes chemotaxis behaviour of mesenchymal stem cells via HIF-1α signalling.
Journal of cellular and molecular medicine
Xu W,Xu R,Li Z,Wang Y,Hu R
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2019
Therapeutic Effects of Human Urine-Derived Stem Cells in a Rat Model of Cisplatin-Induced Acute Kidney Injury In Vivo and In Vitro.
Stem cells international
Sun B,Luo X,Yang C,Liu P,Yang Y,Dong X,Yang Z,Xu J,Zhang Y,Li L
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu May 13 00:00:00 EDT 2021
HMGB3 silence inhibits breast cancer cell proliferation and tumor growth by interacting with hypoxia-inducible factor 1α.
Cancer management and research
Gu J,Xu T,Huang QH,Zhang CM,Chen HY
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2020
TGF‑β induces periodontal ligament stem cell senescence through increase of ROS production.
Molecular medicine reports
Fan C,Ji Q,Zhang C,Xu S,Sun H,Li Z
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2019
Effects of demographic factors on adipogenic and chondrogenic differentiation in bone marrow-derived stem cells.
Experimental and therapeutic medicine
Lee H,Min SK,Park JB
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed May 01 00:00:00 EDT 2019
High-yield isolation of menstrual blood-derived endometrial stem cells by direct red blood cell lysis treatment.
Biology open
Sun Y,Ren Y,Yang F,He Y,Liang S,Guan L,Cheng F,Liu Y,Lin J
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu May 02 00:00:00 EDT 2019
The deubiquitinating enzyme PSMD14 facilitates tumor growth and chemoresistance through stabilizing the ALK2 receptor in the initiation of BMP6 signaling pathway.
EBioMedicine
Seo D,Jung SM,Park JS,Lee J,Ha J,Kim M,Park SH
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Nov 01 00:00:00 EDT 2019
Overexpression of Aiolos promotes epithelial-mesenchymal transition and cancer stem cell-like properties in lung cancer cells.
Scientific reports
Hung JJ,Kao YS,Huang CH,Hsu WH
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Feb 28 00:00:00 EST 2019
BMX-ARHGAP fusion protein maintains the tumorigenicity of gastric cancer stem cells by activating the JAK/STAT3 signaling pathway.
Cancer cell international
Xu XF,Gao F,Wang JJ,Long C,Chen X,Tao L,Yang L,Ding L,Ji Y
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Apr 08 00:00:00 EDT 2022
Sex Differences in Mouse Popliteal Lymph Nodes.
Scientific reports
Dill-Garlow R,Chen KE,Walker AM
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Jan 30 00:00:00 EST 2019
Sex Differences in Mouse Popliteal Lymph Nodes.
Scientific reports
Dill-Garlow R,Chen KE,Walker AM
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Jan 30 00:00:00 EST 2019
ER stress-induced mediator C/EBP homologous protein thwarts effector T cell activity in tumors through T-bet repression.
Nature communications
Cao Y,Trillo-Tinoco J,Sierra RA,Anadon C,Dai W,Mohamed E,Cen L,Costich TL,Magliocco A,Marchion D,Klar R,Michel S,Jaschinski F,Reich RR,Mehrotra S,Cubillos-Ruiz JR,Munn DH,Conejo-Garcia JR,Rodriguez PC
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Mar 20 00:00:00 EDT 2019
Human Pluripotent Stem Cell-Derived Multipotent Vascular Progenitors of the Mesothelium Lineage Have Utility in Tissue Engineering and Repair.
Cell reports
Colunga T,Hayworth M,Kreß S,Reynolds DM,Chen L,Nazor KL,Baur J,Singh AM,Loring JF,Metzger M,Dalton S
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Mar 05 00:00:00 EST 2019
IL-17 inhibits CXCL9/10-mediated recruitment of CD8+ cytotoxic T cells and regulatory T cells to colorectal tumors.
Journal for immunotherapy of cancer
Chen J,Ye X,Pitmon E,Lu M,Wan J,Jellison ER,Adler AJ,Vella AT,Wang K
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Nov 27 00:00:00 EST 2019
BRCA1 regulates the cancer stem cell fate of breast cancer cells in the context of hypoxia and histone deacetylase inhibitors.
Scientific reports
Kim H,Lin Q,Yun Z
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Jul 04 00:00:00 EDT 2019
Visfatin Mediates Malignant Behaviors through Adipose-Derived Stem Cells Intermediary in Breast Cancer.
Cancers
Huang JY,Wang YY,Lo S,Tseng LM,Chen DR,Wu YC,Hou MF,Yuan SF
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Dec 20 00:00:00 EST 2019
Growth Factor Screening in Dystrophic Muscles Reveals PDGFB/PDGFRB-Mediated Migration of Interstitial Stem Cells.
International journal of molecular sciences
Camps J,Grosemans H,Gijsbers R,Maes C,Sampaolesi M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Mar 05 00:00:00 EST 2019
Conserved regulation of RNA processing in somatic cell reprogramming.
BMC genomics
Kanitz A,Syed AP,Kaji K,Zavolan M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Jan 31 00:00:00 EST 2019
Better therapeutic potential of bone marrow-derived mesenchymal stem cells compared with chorionic villi-derived mesenchymal stem cells in airway injury model.
Regenerative medicine
Ji S,Wu C,Tong L,Wang L,Zhou J,Chen C,Song Y
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2019
Peptide:MHCII Tetramer-Based Cell Enrichment for the Study of Epitope-Specific CD4+ T Cells.
Current protocols in immunology
Kotov DI,Jenkins MK
56-0441 was used in Flow cytometry/Cell sorting to study polyclonal epitope-specific CD4+ T cells under physiological conditions.
Sat Jun 01 00:00:00 EDT 2019
Non-conventional Inhibitory CD4+Foxp3-PD-1hi T Cells as a Biomarker of Immune Checkpoint Blockade Activity.
Cancer cell
Zappasodi R,Budhu S,Hellmann MD,Postow MA,Senbabaoglu Y,Manne S,Gasmi B,Liu C,Zhong H,Li Y,Huang AC,Hirschhorn-Cymerman D,Panageas KS,Wherry EJ,Merghoub T,Wolchok JD
56-0441-82 was used in Flow Cytometry to provide evidence that mouse and human circulating and intra-tumor 4PD1hi inhibit T cell functions in a PD-1/PD-L1 dependent fashion and resemble follicular helper T cell (TFH)-like cells.
Mon Jun 11 00:00:00 EDT 2018
Bryostatin-1 alleviates experimental multiple sclerosis.
Proceedings of the National Academy of Sciences of the United States of America
Kornberg MD,Smith MD,Shirazi HA,Calabresi PA,Snyder SH,Kim PM
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Feb 27 00:00:00 EST 2018
Mutant p53 gain of function underlies high expression levels of colorectal cancer stem cells markers.
Oncogene
Solomon H,Dinowitz N,Pateras IS,Cooks T,Shetzer Y,Molchadsky A,Charni M,Rabani S,Koifman G,Tarcic O,Porat Z,Kogan-Sakin I,Goldfinger N,Oren M,Harris CC,Gorgoulis VG,Rotter V
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Mar 01 00:00:00 EST 2018
The hypoxic tumor microenvironment in vivo selects the cancer stem cell fate of breast cancer cells.
Breast cancer research : BCR
Kim H,Lin Q,Glazer PM,Yun Z
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Mar 06 00:00:00 EST 2018
SUV420H2 is an epigenetic regulator of epithelial/mesenchymal states in pancreatic cancer.
The Journal of cell biology
Viotti M,Wilson C,McCleland M,Koeppen H,Haley B,Jhunjhunwala S,Klijn C,Modrusan Z,Arnott D,Classon M,Stephan JP,Mellman I
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Feb 05 00:00:00 EST 2018
CCR7 on CD4+ T Cells Plays a Crucial Role in the Induction of Experimental Autoimmune Encephalomyelitis.
Journal of immunology (Baltimore, Md. : 1950)
Belikan P,Bühler U,Wolf C,Pramanik GK,Gollan R,Zipp F,Siffrin V
56-0441 was used in Flow cytometry/Cell sorting to indicate a crucial role of CCR7 in neuroinflammation during the priming of autoimmune CD4+ T cells but not in the CNS.
Sun Apr 15 00:00:00 EDT 2018
Polymethoxylated Flavones from Orange Peels Inhibit Cell Proliferation in a 3D Cell Model of Human Colorectal Cancer.
Nutrition and cancer
Silva I,Estrada MF,V Pereira C,da Silva AB,Bronze MR,Alves PM,Duarte CMM,Brito C,Serra AT
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunocytochemistry
Wed Apr 03 00:00:00 EDT 2019
Tanshinone IIA and Astragaloside IV promote the angiogenesis of mesenchymal stem cell-derived endothelial cell-like cells via upregulation of Cx37, Cx40 and Cx43.
Experimental and therapeutic medicine
Li Z,Zhang S,Cao L,Li W,Ye YC,Shi ZX,Wang ZR,Sun LX,Wang JW,Jia LT,Wang W
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Feb 01 00:00:00 EST 2018
LGR5, a novel functional glioma stem cell marker, promotes EMT by activating the Wnt/β-catenin pathway and predicts poor survival of glioma patients.
Journal of experimental & clinical cancer research : CR
Zhang J,Cai H,Sun L,Zhan P,Chen M,Zhang F,Ran Y,Wan J
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunohistochemistry
Wed Sep 12 00:00:00 EDT 2018
mTOR Modulates CD8+ T Cell Differentiation in Mice with Invasive Pulmonary Aspergillosis.
Open life sciences
Wang H,Xiao Y,Su L,Cui N,Liu D
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
Oncofetal gene SALL4 reactivation by hepatitis B virus counteracts miR-200c in PD-L1-induced T cell exhaustion.
Nature communications
Sun C,Lan P,Han Q,Huang M,Zhang Z,Xu G,Song J,Wang J,Wei H,Zhang J,Sun R,Zhang C,Tian Z
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Mar 28 00:00:00 EDT 2018
Inhibition of Fas associated phosphatase 1 (Fap1) facilitates apoptosis of colon cancer stem cells and enhances the effects of oxaliplatin.
Oncotarget
Huang W,Bei L,Eklund EA
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri May 25 00:00:00 EDT 2018
An alternative splicing switch in FLNB promotes the mesenchymal cell state in human breast cancer.
eLife
Li J,Choi PS,Chaffer CL,Labella K,Hwang JH,Giacomelli AO,Kim JW,Ilic N,Doench JG,Ly SH,Dai C,Hagel K,Hong AL,Gjoerup O,Goel S,Ge JY,Root DE,Zhao JJ,Brooks AN,Weinberg RA,Hahn WC
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Jul 30 00:00:00 EDT 2018
Modifying the cancer-immune set point using vaccinia virus expressing re-designed interleukin-2.
Nature communications
Liu Z,Ge Y,Wang H,Ma C,Feist M,Ju S,Guo ZS,Bartlett DL
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Nov 08 00:00:00 EST 2018
RhoA, Rac1, and Cdc42 differentially regulate αSMA and collagen I expression in mesenchymal stem cells.
The Journal of biological chemistry
Ge J,Burnier L,Adamopoulou M,Kwa MQ,Schaks M,Rottner K,Brakebusch C
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Jun 15 00:00:00 EDT 2018
Selective Export into Extracellular Vesicles and Function of tRNA Fragments during T Cell Activation.
Cell reports
Chiou NT,Kageyama R,Ansel KM
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Dec 18 00:00:00 EST 2018
Isolation and characterization of mesenchymal stem/stromal cells from Ctenomys minutus.
Genetics and molecular biology
Pereira MRJ,Pinhatti VR,Silveira MDD,Matzenbacher CA,Freitas TRO,Silva JD,Camassola M,Nardi NB
56-0441-82 was used in Flow cytometry/Cell sorting to show that the collection of biological information for biobanks represents an important contribution to the creation of strategies for prevention of loss of genetic diversity.
Thu Oct 01 00:00:00 EDT 2020
Ablation of Transcription Factor IRF4 Promotes Transplant Acceptance by Driving Allogenic CD4+ T Cell Dysfunction.
Immunity
Wu J,Zhang H,Shi X,Xiao X,Fan Y,Minze LJ,Wang J,Ghobrial RM,Xia J,Sciammas R,Li XC,Chen W
56-0441-82 was used in Flow Cytometry to demonstrate that interferon regulatory factor 4 was a key transcriptional determinant controlling T cell responses during transplantation.
Tue Dec 19 00:00:00 EST 2017
Antibody Tumor Targeting Is Enhanced by CD27 Agonists through Myeloid Recruitment.
Cancer cell
Turaj AH,Hussain K,Cox KL,Rose-Zerilli MJJ,Testa J,Dahal LN,Chan HTC,James S,Field VL,Carter MJ,Kim HJ,West JJ,Thomas LJ,He LZ,Keler T,Johnson PWM,Al-Shamkhani A,Thirdborough SM,Beers SA,Cragg MS,Glennie MJ,Lim SH
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Dec 11 00:00:00 EST 2017
Constitutively Active SMAD2/3 Are Broad-Scope Potentiators of Transcription-Factor-Mediated Cellular Reprogramming.
Cell stem cell
Ruetz T,Pfisterer U,Di Stefano B,Ashmore J,Beniazza M,Tian TV,Kaemena DF,Tosti L,Tan W,Manning JR,Chantzoura E,Ottosson DR,Collombet S,Johnsson A,Cohen E,Yusa K,Linnarsson S,Graf T,Parmar M,Kaji K
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Dec 07 00:00:00 EST 2017
Excessive expression of miR-27 impairs Treg-mediated immunological tolerance.
The Journal of clinical investigation
Cruz LO,Hashemifar SS,Wu CJ,Cho S,Nguyen DT,Lin LL,Khan AA,Lu LF
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2017
Egr2 and 3 control adaptive immune responses by temporally uncoupling expansion from T cell differentiation.
The Journal of experimental medicine
Miao T,Symonds ALJ,Singh R,Symonds JD,Ogbe A,Omodho B,Zhu B,Li S,Wang P
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Jun 05 00:00:00 EDT 2017
Integrin-β4 identifies cancer stem cell-enriched populations of partially mesenchymal carcinoma cells.
Proceedings of the National Academy of Sciences of the United States of America
Bierie B,Pierce SE,Kroeger C,Stover DG,Pattabiraman DR,Thiru P,Liu Donaher J,Reinhardt F,Chaffer CL,Keckesova Z,Weinberg RA
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Mar 21 00:00:00 EDT 2017
Role of Triggering Receptor Expressed on Myeloid Cell-1 Expression in Mammalian Target of Rapamycin Modulation of CD8+ T-cell Differentiation during the Immune Response to Invasive Pulmonary Aspergillosis.
Chinese medical journal
Cui N,Wang H,Su LX,Zhang JH,Long Y,Liu DW
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sat May 20 00:00:00 EDT 2017
The Ox40/Ox40 Ligand Pathway Promotes Pathogenic Th Cell Responses, Plasmablast Accumulation, and Lupus Nephritis in NZB/W F1 Mice.
Journal of immunology (Baltimore, Md. : 1950)
Sitrin J,Suto E,Wuster A,Eastham-Anderson J,Kim JM,Austin CD,Lee WP,Behrens TW
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Aug 15 00:00:00 EDT 2017
Chemokine Receptor-Dependent Control of Skin Tissue-Resident Memory T Cell Formation.
Journal of immunology (Baltimore, Md. : 1950)
Zaid A,Hor JL,Christo SN,Groom JR,Heath WR,Mackay LK,Mueller SN
56-0441 was used in Flow cytometry/Cell sorting to examine the signals required for the migration of tissue-resident memory T cells within the constrained epidermal environment.
Sun Oct 01 00:00:00 EDT 2017
A high-yield isolation and enrichment strategy for human lung microvascular endothelial cells.
Pulmonary circulation
Gaskill C,Majka SM
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2017
Comparison of Four Protocols to Generate Chondrocyte-Like Cells from Human Induced Pluripotent Stem Cells (hiPSCs).
Stem cell reviews and reports
Suchorska WM,Augustyniak E,Richter M,Trzeciak T
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
Targeted genome editing restores T cell differentiation in a humanized X-SCID pluripotent stem cell disease model.
Scientific reports
Alzubi J,Pallant C,Mussolino C,Howe SJ,Thrasher AJ,Cathomen T
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Sep 29 00:00:00 EDT 2017
Tristetraprolin inhibits macrophage IL-27-induced activation of antitumour cytotoxic T cell responses.
Nature communications
Wang Q,Ning H,Peng H,Wei L,Hou R,Hoft DF,Liu J
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Oct 11 00:00:00 EDT 2017
Glutamic Pyruvate Transaminase GPT2 Promotes Tumorigenesis of Breast Cancer Cells by Activating Sonic Hedgehog Signaling.
Theranostics
Cao Y,Lin SH,Wang Y,Chin YE,Kang L,Mi J
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Apr 30 00:00:00 EDT 2018
IL-7-dependent STAT1 activation limits homeostatic CD4+ T cell expansion.
JCI insight
Le Saout C,Luckey MA,Villarino AV,Smith M,Hasley RB,Myers TG,Imamichi H,Park JH,O'Shea JJ,Lane HC,Catalfamo M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Nov 16 00:00:00 EST 2017
SOCS3 treatment prevents the development of alopecia areata by inhibiting CD8+ T cell-mediated autoimmune destruction.
Oncotarget
Gao Z,Jin YQ,Wu W
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue May 16 00:00:00 EDT 2017
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Oct 17 00:00:00 EDT 2017
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Oct 17 00:00:00 EDT 2017
Zoledronate suppressed angiogenesis and osteogenesis by inhibiting osteoclasts formation and secretion of PDGF-BB.
PloS one
Gao SY,Zheng GS,Wang L,Liang YJ,Zhang SE,Lao XM,Li K,Liao GQ
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Sep 18 00:00:00 EDT 2017
Oligodendrocyte death results in immune-mediated CNS demyelination.
Nature neuroscience
Traka M,Podojil JR,McCarthy DP,Miller SD,Popko B
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2016
SOX2 and PI3K Cooperate to Induce and Stabilize a Squamous-Committed Stem Cell Injury State during Lung Squamous Cell Carcinoma Pathogenesis.
PLoS biology
Kim BR,Van de Laar E,Cabanero M,Tarumi S,Hasenoeder S,Wang D,Virtanen C,Suzuki T,Bandarchi B,Sakashita S,Pham NA,Lee S,Keshavjee S,Waddell TK,Tsao MS,Moghal N
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2016
Stem Cells Antigen-1 Enriches for a Cancer Stem Cell-Like Subpopulation in Mouse Gastric Cancer.
Stem cells (Dayton, Ohio)
Park JW,Park JM,Park DM,Kim DY,Kim HK
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sun May 01 00:00:00 EDT 2016
Resident T Cells Are Unable To Control Herpes Simplex Virus-1 Activity in the Brain Ependymal Region during Latency.
Journal of immunology (Baltimore, Md. : 1950)
Menendez CM,Jinkins JK,Carr DJ
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Aug 15 00:00:00 EDT 2016
Lunatic, Manic, and Radical Fringe Each Promote T and B Cell Development.
Journal of immunology (Baltimore, Md. : 1950)
Song Y,Kumar V,Wei HX,Qiu J,Stanley P
56-0441 was used in Flow cytometry/Cell sorting to investigate the roles for maniac fringe (MFNG) and radical fringe (RFNG) in T cell development, RFNG in B cell development, and Fringes in T and B cell activation, showing that they promote T and B cell development.
Fri Jan 01 00:00:00 EST 2016
Helios Controls a Limited Subset of Regulatory T Cell Functions.
Journal of immunology (Baltimore, Md. : 1950)
Sebastian M,Lopez-Ocasio M,Metidji A,Rieder SA,Shevach EM,Thornton AM
56-0441 was used in Flow cytometry/Cell sorting to determine the beneficial effects of the transcription factor, Helios, for Treg maintenance, differentiation, and survival.
Fri Jan 01 00:00:00 EST 2016
Effects of nerve growth factor and basic fibroblast growth factor dual gene modification on rat bone marrow mesenchymal stem cell differentiation into neuron-like cells in vitro.
Molecular medicine reports
Hu Y,Zhang Y,Tian K,Xun C,Wang S,Lv D
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2016
Breast cancer stem cells expressing different stem cell markers exhibit distinct biological characteristics.
Molecular medicine reports
Shao J,Fan W,Ma B,Wu Y
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunofluorescence
Thu Dec 01 00:00:00 EST 2016
Comparable roles of CD44v8-10 and CD44s in the development of bone metastases in a mouse model.
Oncology letters
Hiraga T,Nakamura H
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sat Oct 01 00:00:00 EDT 2016
Cryptotanshinone targets tumor-initiating cells through down-regulation of stemness genes expression.
Oncology letters
Zhang Y,Cabarcas SM,Zheng JI,Sun L,Mathews LA,Zhang X,Lin H,Farrar WL
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2016
PRDM14 promotes RAG-dependent Notch1 driver mutations in mouse T-ALL.
Biology open
Carofino BL,Ayanga B,Tracey LJ,Brooke-Bisschop T,Justice MJ
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sun May 15 00:00:00 EDT 2016
Sertoli cell condition medium can induce germ like cells from bone marrow derived mesenchymal stem cells.
Iranian journal of basic medical sciences
Monfared MH,Minaee B,Rastegar T,Khrazinejad E,Barbarestani M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2016
A cell-autonomous tumour suppressor role of RAF1 in hepatocarcinogenesis.
Nature communications
Jeric I,Maurer G,Cavallo AL,Raguz J,Desideri E,Tarkowski B,Parrini M,Fischer I,Zatloukal K,Baccarini M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Dec 21 00:00:00 EST 2016
Treg engage lymphotoxin beta receptor for afferent lymphatic transendothelial migration.
Nature communications
Brinkman CC,Iwami D,Hritzo MK,Xiong Y,Ahmad S,Simon T,Hippen KL,Blazar BR,Bromberg JS
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Jun 21 00:00:00 EDT 2016
The Z-cad dual fluorescent sensor detects dynamic changes between the epithelial and mesenchymal cellular states.
BMC biology
Toneff MJ,Sreekumar A,Tinnirello A,Hollander PD,Habib S,Li S,Ellis MJ,Xin L,Mani SA,Rosen JM
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Jun 17 00:00:00 EDT 2016
Identification and validation of multiple cell surface markers of clinical-grade adipose-derived mesenchymal stromal cells as novel release criteria for good manufacturing practice-compliant production.
Stem cell research & therapy
Camilleri ET,Gustafson MP,Dudakovic A,Riester SM,Garces CG,Paradise CR,Takai H,Karperien M,Cool S,Sampen HJ,Larson AN,Qu W,Smith J,Dietz AB,van Wijnen AJ
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Aug 11 00:00:00 EDT 2016
Equine-Induced Pluripotent Stem Cells Retain Lineage Commitment Toward Myogenic and Chondrogenic Fates.
Stem cell reports
Quattrocelli M,Giacomazzi G,Broeckx SY,Ceelen L,Bolca S,Spaas JH,Sampaolesi M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunofluorescence
Tue Jan 12 00:00:00 EST 2016
Equine-Induced Pluripotent Stem Cells Retain Lineage Commitment Toward Myogenic and Chondrogenic Fates.
Stem cell reports
Quattrocelli M,Giacomazzi G,Broeckx SY,Ceelen L,Bolca S,Spaas JH,Sampaolesi M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunofluorescence
Tue Jan 12 00:00:00 EST 2016
Llgl1 prevents metaplastic survival driven by epidermal growth factor dependent migration.
Oncotarget
Greenwood E,Maisel S,Ebertz D,Russ A,Pandey R,Schroeder J
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Sep 20 00:00:00 EDT 2016
Itk is required for Th9 differentiation via TCR-mediated induction of IL-2 and IRF4.
Nature communications
Gomez-Rodriguez J,Meylan F,Handon R,Hayes ET,Anderson SM,Kirby MR,Siegel RM,Schwartzberg PL
56-0441 was used in Flow cytometry/Cell sorting to investigate the role of IL-9 in allergic asthma and autoimmunity, showing that Itk is required for Th9 differentiation via TCR-mediated induction of IL-2 and IRF4.
Thu Mar 03 00:00:00 EST 2016
Anisotropic stress orients remodelling of mammalian limb bud ectoderm.
Nature cell biology
Lau K,Tao H,Liu H,Wen J,Sturgeon K,Sorfazlian N,Lazic S,Burrows JT,Wong MD,Li D,Deimling S,Ciruna B,Scott I,Simmons C,Henkelman RM,Williams T,Hadjantonakis AK,Fernandez-Gonzalez R,Sun Y,Hopyan S
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunofluorescence
Fri May 01 00:00:00 EDT 2015
Mesodermal iPSC-derived progenitor cells functionally regenerate cardiac and skeletal muscle.
The Journal of clinical investigation
Quattrocelli M,Swinnen M,Giacomazzi G,Camps J,Barthélemy I,Ceccarelli G,Caluwé E,Grosemans H,Thorrez L,Pelizzo G,Muijtjens M,Verfaillie CM,Blot S,Janssens S,Sampaolesi M
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2015
Manic fringe promotes a claudin-low breast cancer phenotype through notch-mediated PIK3CG induction.
Cancer research
Zhang S,Chung WC,Wu G,Egan SE,Miele L,Xu K
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri May 15 00:00:00 EDT 2015
Leptin receptor signaling in T cells is required for Th17 differentiation.
Journal of immunology (Baltimore, Md. : 1950)
Reis BS,Lee K,Fanok MH,Mascaraque C,Amoury M,Cohn LB,Rogoz A,Dallner OS,Moraes-Vieira PM,Domingos AI,Mucida D
56-0441 was used in Flow cytometry/Cell sorting to show that leptin receptor signalling in T cells is required for Th17 differentiation.
Mon Jun 01 00:00:00 EDT 2015
Enrichment of Human Stem-Like Prostate Cells with s-SHIP Promoter Activity Uncovers a Role in Stemness for the Long Noncoding RNA H19.
Stem cells and development
Bauderlique-Le Roy H,Vennin C,Brocqueville G,Spruyt N,Adriaenssens E,Bourette RP
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri May 15 00:00:00 EDT 2015
Multi-Drug Resistance ABC Transporter Inhibition Enhances Murine Ventral Prostate Stem/Progenitor Cell Differentiation.
Stem cells and development
Samant MD,Jackson CM,Felix CL,Jones AJ,Goodrich DW,Foster BA,Huss WJ
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri May 15 00:00:00 EDT 2015
Triclosan Induces Thymic Stromal Lymphopoietin in Skin Promoting Th2 Allergic Responses.
Toxicological sciences : an official journal of the Society of Toxicology
Marshall NB,Lukomska E,Long CM,Kashon ML,Sharpnack DD,Nayak AP,Anderson KL,Jean Meade B,Anderson SE
56-0441 was used in Flow cytometry/Cell sorting to examine the effects of dermal triclosan exposure on the skin and lymph nodes of mice and in a human skin model to identify mechanisms for augmenting allergic responses.
Tue Sep 01 00:00:00 EDT 2015
Special AT-rich sequence-binding protein-1 participates in the maintenance of breast cancer stem cells through regulation of the Notch signaling pathway and expression of Snail1 and Twist1.
Molecular medicine reports
Sun Z,Zhang C,Zou X,Jiang G,Xu Z,Li W,Xie H
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri May 01 00:00:00 EDT 2015
Reprogramming Roadblocks Are System Dependent.
Stem cell reports
Chantzoura E,Skylaki S,Menendez S,Kim SI,Johnsson A,Linnarsson S,Woltjen K,Chambers I,Kaji K
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Sep 08 00:00:00 EDT 2015
Multi-lineage differentiation of human umbilical cord Wharton's Jelly Mesenchymal Stromal Cells mediates changes in the expression profile of stemness markers.
PloS one
Ali H,Al-Yatama MK,Abu-Farha M,Behbehani K,Al Madhoun A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Mar 25 00:00:00 EDT 2016
Multi-lineage differentiation of human umbilical cord Wharton's Jelly Mesenchymal Stromal Cells mediates changes in the expression profile of stemness markers.
PloS one
Ali H,Al-Yatama MK,Abu-Farha M,Behbehani K,Al Madhoun A
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Mar 25 00:00:00 EDT 2016
Human adipose-derived stem cells partially rescue the stroke syndromes by promoting spatial learning and memory in mouse middle cerebral artery occlusion model.
Stem cell research & therapy
Zhou F,Gao S,Wang L,Sun C,Chen L,Yuan P,Zhao H,Yi Y,Qin Y,Dong Z,Cao L,Ren H,Zhu L,Li Q,Lu B,Liang A,Xu GT,Zhu H,Gao Z,Ma J,Xu J,Chen X
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sat May 09 00:00:00 EDT 2015
Prophylaxis with a respiratory syncytial virus (RSV) anti-G protein monoclonal antibody shifts the adaptive immune response to RSV rA2-line19F infection from Th2 to Th1 in BALB/c mice.
Journal of virology
Boyoglu-Barnum S,Chirkova T,Todd SO,Barnum TR,Gaston KA,Jorquera P,Haynes LM,Tripp RA,Moore ML,Anderson LJ
56-0441 was used in Flow cytometry/Cell sorting to suggest that the RSV G protein not only contributes to disease, but also dampens the host immune response to infection.
Mon Sep 01 00:00:00 EDT 2014
Bispecificity for myelin and neuronal self-antigens is a common feature of CD4 T cells in C57BL/6 mice.
Journal of immunology (Baltimore, Md. : 1950)
Lucca LE,Desbois S,Ramadan A,Ben-Nun A,Eisenstein M,Carrié N,Guéry JC,Sette A,Nguyen P,Geiger TL,Mars LT,Liblau RS
56-0441 was used in Flow cytometry/Cell sorting to assess whether cross-reactivity is a common feature of the myelin oligodendrocyte glycoprotein epitope 35-55-specific T cell response.
Wed Oct 01 00:00:00 EDT 2014
An essential role for medullary thymic epithelial cells during the intrathymic development of invariant NKT cells.
Journal of immunology (Baltimore, Md. : 1950)
White AJ,Jenkinson WE,Cowan JE,Parnell SM,Bacon A,Jones ND,Jenkinson EJ,Anderson G
56-0441 was used in Flow cytometry/Cell sorting to investigate whether thymus microenvironments are required in iNKT cell development, showing that medullary thymic epithelial cells are essential for intrathymic development of iNKT cells.
Sat Mar 15 00:00:00 EDT 2014
Differential requirement for CCR4 and CCR7 during the development of innate and adaptive αβT cells in the adult thymus.
Journal of immunology (Baltimore, Md. : 1950)
Cowan JE,McCarthy NI,Parnell SM,White AJ,Bacon A,Serge A,Irla M,Lane PJ,Jenkinson EJ,Jenkinson WE,Anderson G
56-0441 was used in Flow cytometry/Cell sorting to identify novel roles for CCR7 during intrathymic T cell development, highlighting its importance in enabling multiple αβT cell lineages to access the thymic medulla.
Fri Aug 01 00:00:00 EDT 2014
Serum- and growth-factor-free three-dimensional culture system supports cartilage tissue formation by promoting collagen synthesis via Sox9-Col2a1 interaction.
Tissue engineering. Part A
Ahmed N,Iu J,Brown CE,Taylor DW,Kandel RA
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Aug 01 00:00:00 EDT 2014
Tumor-suppressive activity of Lunatic Fringe in prostate through differential modulation of Notch receptor activation.
Neoplasia (New York, N.Y.)
Zhang S,Chung WC,Wu G,Egan SE,Xu K
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Sat Feb 01 00:00:00 EST 2014
FOXF1 mediates mesenchymal stem cell fusion-induced reprogramming of lung cancer cells.
Oncotarget
Wei HJ,Nickoloff JA,Chen WH,Liu HY,Lo WC,Chang YT,Yang PC,Wu CW,Williams DF,Gelovani JG,Deng WP
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Wed Oct 15 00:00:00 EDT 2014
Amniotic fluid stem cells prevent follicle atresia and rescue fertility of mice with premature ovarian failure induced by chemotherapy.
PloS one
Xiao GY,Liu IH,Cheng CC,Chang CC,Lee YH,Cheng WT,Wu SC
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu May 28 00:00:00 EDT 2015
Validation of the effects of TGF-β1 on tumor recurrence and prognosis through tumor retrieval and cell mechanical properties.
Cancer cell international
Wu TH,Chou YW,Chiu PH,Tang MJ,Hu CW,Yeh ML
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Mar 03 00:00:00 EST 2014
High-resolution analysis with novel cell-surface markers identifies routes to iPS cells.
Nature
O'Malley J,Skylaki S,Iwabuchi KA,Chantzoura E,Ruetz T,Johnsson A,Tomlinson SR,Linnarsson S,Kaji K
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Jul 04 00:00:00 EDT 2013
Mutual expression of the transcription factors Runx3 and ThPOK regulates intestinal CD4⁺ T cell immunity.
Nature immunology
Reis BS,Rogoz A,Costa-Pinto FA,Taniuchi I,Mucida D
56-0441 was used in Flow cytometry/Cell sorting to demonstrate plasticity in the CD4(+) T cell lineage that allows chronic exposure to luminal antigens without pathological inflammation.
Fri Mar 01 00:00:00 EST 2013
miR-126 and miR-126* repress recruitment of mesenchymal stem cells and inflammatory monocytes to inhibit breast cancer metastasis.
Nature cell biology
Zhang Y,Yang P,Sun T,Li D,Xu X,Rui Y,Li C,Chong M,Ibrahim T,Mercatali L,Amadori D,Lu X,Xie D,Li QJ,Wang XF
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Feb 01 00:00:00 EST 2013
The thymic medulla is required for Foxp3+ regulatory but not conventional CD4+ thymocyte development.
The Journal of experimental medicine
Cowan JE,Parnell SM,Nakamura K,Caamano JH,Lane PJ,Jenkinson EJ,Jenkinson WE,Anderson G
56-0441 was used in Flow cytometry/Cell sorting to demonstrate a differential requirement for the thymic medulla in relation to CD4 conventional and Foxp3(+) thymocyte lineages.
Mon Apr 08 00:00:00 EDT 2013
Inhibiting interactions of lysine demethylase LSD1 with snail/slug blocks cancer cell invasion.
Cancer research
Ferrari-Amorotti G,Fragliasso V,Esteki R,Prudente Z,Soliera AR,Cattelani S,Manzotti G,Grisendi G,Dominici M,Pieraccioli M,Raschellà G,Chiodoni C,Colombo MP,Calabretta B
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Jan 01 00:00:00 EST 2013
Evidence for a multipotent mammary progenitor with pregnancy-specific activity.
Breast cancer research : BCR
Kaanta AS,Virtanen C,Selfors LM,Brugge JS,Neel BG
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Dec 08 00:00:00 EST 2014
Phenotypic conversions of "protoplasmic" to "reactive" astrocytes in Alexander disease.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Sosunov AA,Guilfoyle E,Wu X,McKhann GM,Goldman JE
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunofluorescence
Wed Apr 24 00:00:00 EDT 2013
The CD44+ ALDH+ population of human keratinocytes is enriched for epidermal stem cells with long-term repopulating ability.
Stem cells (Dayton, Ohio)
Szabo AZ,Fong S,Yue L,Zhang K,Strachan LR,Scalapino K,Mancianti ML,Ghadially R
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Apr 01 00:00:00 EDT 2013
The CD44+ ALDH+ population of human keratinocytes is enriched for epidermal stem cells with long-term repopulating ability.
Stem cells (Dayton, Ohio)
Szabo AZ,Fong S,Yue L,Zhang K,Strachan LR,Scalapino K,Mancianti ML,Ghadially R
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Apr 01 00:00:00 EDT 2013
The CD44+ ALDH+ population of human keratinocytes is enriched for epidermal stem cells with long-term repopulating ability.
Stem cells (Dayton, Ohio)
Szabo AZ,Fong S,Yue L,Zhang K,Strachan LR,Scalapino K,Mancianti ML,Ghadially R
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Apr 01 00:00:00 EDT 2013
Estrogen and progesterone together expand murine endometrial epithelial progenitor cells.
Stem cells (Dayton, Ohio)
Janzen DM,Cheng D,Schafenacker AM,Paik DY,Goldstein AS,Witte ON,Jaroszewicz A,Pellegrini M,Memarzadeh S
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Apr 01 00:00:00 EDT 2013
Estrogen and progesterone together expand murine endometrial epithelial progenitor cells.
Stem cells (Dayton, Ohio)
Janzen DM,Cheng D,Schafenacker AM,Paik DY,Goldstein AS,Witte ON,Jaroszewicz A,Pellegrini M,Memarzadeh S
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Apr 01 00:00:00 EDT 2013
Increased invasion and tumorigenicity capacity of CD44+/CD24- breast cancer MCF7 cells in vitro and in nude mice.
Cancer cell international
Yan W,Chen Y,Yao Y,Zhang H,Wang T
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Mon Jun 24 00:00:00 EDT 2013
Microvesicles derived from human umbilical cord Wharton's jelly mesenchymal stem cells attenuate bladder tumor cell growth in vitro and in vivo.
PloS one
Wu S,Ju GQ,Du T,Zhu YJ,Liu GH
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue Oct 22 00:00:00 EDT 2013
Increased protection from vaccinia virus infection in mice genetically prone to lymphoproliferative disorders.
Journal of virology
Seedhom MO,Mathurin KS,Kim SK,Welsh RM
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2012
Stem-like epithelial cells are concentrated in the distal end of the fallopian tube: a site for injury and serous cancer initiation.
Stem cells (Dayton, Ohio)
Paik DY,Janzen DM,Schafenacker AM,Velasco VS,Shung MS,Cheng D,Huang J,Witte ON,Memarzadeh S
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunohistochemistry
Thu Nov 01 00:00:00 EDT 2012
Stem-like epithelial cells are concentrated in the distal end of the fallopian tube: a site for injury and serous cancer initiation.
Stem cells (Dayton, Ohio)
Paik DY,Janzen DM,Schafenacker AM,Velasco VS,Shung MS,Cheng D,Huang J,Witte ON,Memarzadeh S
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunohistochemistry
Thu Nov 01 00:00:00 EDT 2012
Requirement for diverse TCR specificities determines regulatory T cell activity in a mouse model of autoimmune arthritis.
Journal of immunology (Baltimore, Md. : 1950)
Oh S,Aitken M,Simons DM,Basehoar A,Garcia V,Kropf E,Caton AJ
56-0441 was used in Flow cytometry/Cell sorting to investigate why CD4(+)CD25(+)Foxp3(+) Tregs allow organ-specific autoimmunity, showing that polyclonal Tregs suppressed Th17 cell formation and prevented arthritis development, but clonotypic Tregs did not, indicating that diverse TCR specificities are required.
Tue May 01 00:00:00 EDT 2012
Arthritogenic self-reactive CD4+ T cells acquire an FR4hiCD73hi anergic state in the presence of Foxp3+ regulatory T cells.
Journal of immunology (Baltimore, Md. : 1950)
Martinez RJ,Zhang N,Thomas SR,Nandiwada SL,Jenkins MK,Binstadt BA,Mueller DL
56-0441 was used in Flow cytometry/Cell sorting to suggest that autoimmune arthritis develops in the setting of lymphopenia when Foxp3(+)CD4(+) regulatory T cells are insufficient to functionally inactivate all autoreactive CD4(+) T cells that encounter self-Ag.
Sun Jan 01 00:00:00 EST 2012
The high and low molecular weight forms of hyaluronan have distinct effects on CD44 clustering.
The Journal of biological chemistry
Yang C,Cao M,Liu H,He Y,Xu J,Du Y,Liu Y,Wang W,Cui L,Hu J,Gao F
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Fluorescence Resonance Energy Transfer (FRET)
Fri Dec 14 00:00:00 EST 2012
The high and low molecular weight forms of hyaluronan have distinct effects on CD44 clustering.
The Journal of biological chemistry
Yang C,Cao M,Liu H,He Y,Xu J,Du Y,Liu Y,Wang W,Cui L,Hu J,Gao F
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Fluorescence Resonance Energy Transfer (FRET)
Fri Dec 14 00:00:00 EST 2012
Memory phenotype CD4 T cells undergoing rapid, nonburst-like, cytokine-driven proliferation can be distinguished from antigen-experienced memory cells.
PLoS biology
Younes SA,Punkosdy G,Caucheteux S,Chen T,Grossman Z,Paul WE
56-0441 was used in Flow cytometry/Cell sorting to identify and characterise two sub-populations of memory T cells.
Sat Oct 01 00:00:00 EDT 2011
Increased apoptosis, curtailed expansion and incomplete differentiation of CD8+ T cells combine to decrease clearance of L. monocytogenes in old mice.
European journal of immunology
Smithey MJ,Renkema KR,Rudd BD,Nikolich-Žugich J
56-0441 was used in Flow cytometry/Cell sorting to show that C57BL/6 (B6) mice exhibit an age-dependent reduction in survival, with delayed bacterial clearance in old animals.
Sun May 01 00:00:00 EDT 2011
GLI1 confers profound phenotypic changes upon LNCaP prostate cancer cells that include the acquisition of a hormone independent state.
PloS one
Nadendla SK,Hazan A,Ward M,Harper LJ,Moutasim K,Bianchi LS,Naase M,Ghali L,Thomas GJ,Prowse DM,Philpott MP,Neill GW
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Sep 30 00:00:00 EDT 2011
PTEN loss accelerates KrasG12D-induced pancreatic cancer development.
Cancer research
Hill R,Calvopina JH,Kim C,Wang Y,Dawson DW,Donahue TR,Dry S,Wu H
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunohistochemistry
Wed Sep 15 00:00:00 EDT 2010
CD44 expression is developmentally regulated in the mouse lens and increases in the lens epithelium after injury.
Differentiation; research in biological diversity
Desai VD,Wang Y,Simirskii VN,Duncan MK
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Immunofluorescence
Mon Feb 01 00:00:00 EST 2010
iNKT cell development is orchestrated by different branches of TGF-beta signaling.
The Journal of experimental medicine
Doisne JM,Bartholin L,Yan KP,Garcia CN,Duarte N,Le Luduec JB,Vincent D,Cyprian F,Horvat B,Martel S,Rimokh R,Losson R,Benlagha K,Marie JC
56-0441 was used in Flow cytometry/Cell sorting to show that TGF-beta controls the differentiation program of iNKT cells.
Mon Jun 08 00:00:00 EDT 2009
Tracking epitope-specific T cells.
Nature protocols
Moon JJ,Chu HH,Hataye J,Pagán AJ,Pepper M,McLachlan JB,Zell T,Jenkins MK
56-0441 was used in Flow cytometry/Cell sorting to track epitope-specific T cells.
Tue Jun 30 00:00:00 EDT 2009
Two host factors regulate persistence of H7-specific T cells injected in tumor-bearing mice.
PloS one
Meunier MC,Baron C,Perreault C
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Fri Feb 13 00:00:00 EST 2009
Effects of Culturing on the Stability of the Putative Murine Adipose Derived Stem Cells Markers.
The open stem cell journal
Maddox JR,Liao X,Li F,Niyibizi C
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Thu Jan 01 00:00:00 EST 2009
Modulation of naive CD4+ T-cell responses to an airway antigen during pulmonary mycobacterial infection.
Infection and immunity
Anis MM,Fulton SA,Reba SM,Harding CV,Boom WH
Published figure using CD44 monoclonal antibody (Product # 56-0441-82) in Flow Cytometry
Tue May 01 00:00:00 EDT 2007