Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • CD45 Antibodies

            Invitrogen

            CD45 Monoclonal Antibody (30-F11), eBioscience™

            Advanced Verification
            95 Published Figures
            449 References
            View all (443) CD45 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # 14-0451-82
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (EUR)
            118,00
             
            Check your price
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite CD45 Monoclonal Antibody (30-F11), eBioscience™

            • Antibody Testing Data (1)
            • Published Figures (95)
            • Advanced Verification (1)
            Application
            CD45 Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 97

            CD45 Antibody (14-0451-82) in Flow

            Staining of C57Bl/6 splenocytes with 0.25 µg of Rat IgG2b Isotype Control Purified (Product # 14-4031-82) (open histogram) or 0.25 µg of Anti-Mouse CD45 Purified (filled histogram) followed by Anti-Rat IgG FITC (Product # 11-4811-85). Cells in the lymphocyte gate were used for analysis.
            View Product

            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunocytochemistry (ICC/IF)
            CD45 Antibody in Immunocytochemistry (ICC/IF)
            CD45 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
            CD45 Antibody in Immunocytochemistry (ICC/IF)
            CD45 Antibody in Immunocytochemistry (ICC/IF)
            CD45 Antibody in Immunocytochemistry (ICC/IF)
            CD45 Antibody in Immunocytochemistry (ICC/IF)
            CD45 Antibody in Immunocytochemistry, Immunohistochemistry (Paraffin) (ICC/IF, IHC (P))
            CD45 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
            CD45 Antibody in Immunocytochemistry (ICC/IF)
            CD45 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CD45 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CD45 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CD45 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CD45 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CD45 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CD45 Antibody in Immunohistochemistry (Frozen) (IHC (F))
            CD45 Antibody in Immunohistochemistry (Frozen) (IHC (F))
            CD45 Antibody in Immunohistochemistry (Frozen) (IHC (F))
            CD45 Antibody in Immunohistochemistry (Frozen) (IHC (F))
            CD45 Antibody in Immunohistochemistry (Frozen) (IHC (F))
            CD45 Antibody in Immunohistochemistry (Frozen) (IHC (F))
            CD45 Antibody in Immunohistochemistry (Frozen) (IHC (F))
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (IHC)
            CD45 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
            CD45 Antibody in Western Blot (WB)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody in Flow Cytometry (Flow)
            CD45 Antibody

            Product Details

            14-0451-82

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            -
            View 1 publication 1 publication

            Immunohistochemistry (IHC)

            Assay-Dependent
            View 64 publications 64 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            -
            View 7 publications 7 publications

            Immunohistochemistry (PFA fixed) (IHC (PFA))

            -
            View 2 publications 2 publications

            Immunohistochemistry (Frozen) (IHC (F))

            -
            View 14 publications 14 publications

            Immunocytochemistry (ICC/IF)

            -
            View 16 publications 16 publications

            Flow Cytometry (Flow)

            0.5 µg/test
            View 339 publications 339 publications

            ELISA (ELISA)

            -
            View 1 publication 1 publication

            Immunoprecipitation (IP)

            Assay-Dependent
            -

            Functional Assay (Functional)

            Assay-Dependent
            View 3 publications 3 publications

            In vitro Assay (IV)

            -
            View 1 publication 1 publication

            Miscellaneous PubMed (Misc)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Mouse

            Published species

            Human, Mouse

            Host/Isotype

            Rat / IgG2b, kappa

            Class

            Monoclonal

            Type

            Antibody

            Clone

            30-F11

            Conjugate

            Unconjugated
            • Alexa Fluor 488 
            • Alexa Fluor 532 
            • Alexa Fluor 561
            • Alexa Fluor 660 
            • Alexa Fluor 700 
            • APC 
            • APC-eFluor 780
            • Biotin 
            • Brilliant UV 395
            • Brilliant UV 496
            • Brilliant UV 563
            • Brilliant UV 615
            • Brilliant UV 661
            • Brilliant UV 737
            • Brilliant UV 805
            • Brilliant Violet 421
            • Brilliant Violet 480
            • Brilliant Violet 605
            • Brilliant Violet 650
            • Brilliant Violet 711
            • Brilliant Violet 786
            • eFluor 450
            • eFluor 506
            • FITC
            • Functional Grade
            • NovaFluor Blue 510
            • NovaFluor Blue 555
            • NovaFluor Blue 585
            • NovaFluor Blue 610-30S
            • NovaFluor Blue 610-70S
            • NovaFluor Blue 660-120S
            • NovaFluor Blue 660-40S
            • NovaFluor Blue 725
            • NovaFluor Blue 760
            • NovaFluor Blue 800
            • NovaFluor Red 700
            • NovaFluor Red 710
            • NovaFluor UV 765
            • NovaFluor Violet 690
            • NovaFluor Violet 800
            • NovaFluor Yellow 610
            • NovaFluor Yellow 660
            • NovaFluor Yellow 690
            • NovaFluor Yellow 700
            • NovaFluor Yellow 730
            • NovaFluor Yellow 810
            • PE 
            • PE-Cyanine5
            • PE-Cyanine5.5
            • PE-Cyanine7
            • PE-eFluor 610
            • PerCP-Cyanine5.5
            • Super Bright 436
            • Super Bright 600
            • Super Bright 645
            • Super Bright 702
            • Super Bright 780
            • Request custom conjugation

            Form

            Liquid

            Concentration

            0.5 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2

            Contains

            0.09% sodium azide

            Storage conditions

            4°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_467251

            Product Specific Information

            Description: The 30-F11 monoclonal antibody reacts with all isoforms of mouse CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding mature erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.

            Applications Reported: The 30-F11 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining. 30-F11 has also been reported in complement-dependent cytotoxicity. (Please use Functional Grade purified 30-F11 in functional assays).

            Applications Tested: The 30-F11 antibody has been tested by flow cytometric analysis of mouse bone marrow cells and splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

            Purity: Greater than 90%, as determined by SDS-PAGE.

            Aggregation: Less than 10%, as determined by HPLC.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase (PTP) ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 is absent on non-hematopoietic cell lines, normal and malignant, non-hematopoietic tissues. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing, differ in their extracellular domains but share identical transmembrane and cytoplasmic domains. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45 has been shown to be an essential regulator of T- and B-cell antigen receptor signaling and suppresses JAK kinases to regulate cytokine receptor signaling. CD45 is also important in promoting cell survival by modulating integrin-mediated signal transduction pathway, DNA fragmentation during apoptosis and inhibition or upregulation of various immunological functions.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Related Products for

            Immunohistochemistry
            • Secondary Antibodies
            • Additional Formats
            • Isotype Controls
            • Mounting Media IHC
            • Counter Stains IHC
            • Blockers IHC
            • Substrates IHC
            • Labeling Kits IHC

            Bioinformatics

            Protein Aliases: CD45; L-CA; Leukocyte common antigen; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Receptor-type tyrosine-protein phosphatase C; T200; T220 and B220

            View more View less

            Gene Aliases: B220; Cd45; CD45R; L-CA; loc; Ly-5; Lyt-4; Ptprc; T200

            View more View less

            UniProt ID: (Mouse) P06800

            View more View less

            Entrez Gene ID: (Mouse) 19264

            View more View less

            Function(s)

            phosphoprotein phosphatase activity protein tyrosine phosphatase activity protein binding heparin binding hydrolase activity phosphatase activity protein kinase binding heparan sulfate proteoglycan binding protein phosphatase protein modifying enzyme

            Process(es)

            activation of MAPK activity negative regulation of T cell mediated cytotoxicity positive regulation of T cell mediated cytotoxicity negative regulation of cytokine-mediated signaling pathway hematopoietic progenitor cell differentiation immunoglobulin biosynthetic process positive regulation of T cell mediated immunity regulation of humoral immune response mediated by circulating immunoglobulin positive regulation of humoral immune response mediated by circulating immunoglobulin negative regulation of protein kinase activity protein dephosphorylation negative regulation of cell adhesion involved in substrate-bound cell migration leukocyte cell-cell adhesion response to gamma radiation dephosphorylation B cell differentiation T cell differentiation positive regulation of B cell proliferation negative regulation of protein autophosphorylation heterotypic cell-cell adhesion peptidyl-tyrosine dephosphorylation T cell proliferation B cell proliferation positive regulation of T cell proliferation positive regulation of MAPK cascade cell cycle phase transition positive thymic T cell selection negative thymic T cell selection regulation of B cell differentiation positive regulation of T cell differentiation positive regulation of gamma-delta T cell differentiation positive regulation of alpha-beta T cell proliferation positive regulation of isotype switching to IgG isotypes bone marrow development stem cell development negative regulation of peptidyl-tyrosine phosphorylation T cell receptor signaling pathway B cell receptor signaling pathway regulation of B cell receptor signaling pathway positive regulation of antigen receptor-mediated signaling pathway release of sequestered calcium ion into cytosol defense response to virus regulation of cell cycle positive regulation of hematopoietic stem cell migration positive regulation of stem cell proliferation regulation of extrinsic apoptotic signaling pathway positive regulation of extrinsic apoptotic signaling pathway
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            germany | deutschland flag icon
            Germany | Deutschland

            TEST

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-8vzxs:80/100.66.77.247:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0