Product References
Fibroblastic reticular cells in lymph node potentiate white adipose tissue beiging through neuro-immune crosstalk in male mice.
Nature communications
Cheong LY,Wang B,Wang Q,Jin L,Kwok KHM,Wu X,Shu L,Lin H,Chung SK,Cheng KKY,Hoo RLC,Xu A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Mar 03 00:00:00 EST 2023
Macrophagic AMPKα1 orchestrates regenerative inflammation induced by glucocorticoids.
EMBO reports
Caratti G,Desgeorges T,Juban G,Stifel U,Fessard A,Koenen M,Caratti B,Théret M,Skurk C,Chazaud B,Tuckermann JP,Mounier R
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Feb 06 00:00:00 EST 2023
Topical Diacerein Decreases Skin and Splenic CD11c+ Dendritic Cells in Psoriasis.
International journal of molecular sciences
Brunner SM,Ramspacher A,Rieser C,Leitner J,Heil H,Ablinger M,Tevini J,Wimmer M,Koller A,Piñón Hofbauer J,Felder TK,Bauer JW,Kofler B,Lang R,Wally V
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Feb 21 00:00:00 EST 2023
BRCA1 deficiency in mature CD8+ T lymphocytes impairs antitumor immunity.
Journal for immunotherapy of cancer
Wu B,Qi L,Chiang HC,Pan H,Zhang X,Greenbaum A,Stark E,Wang LJ,Chen Y,Haddad BR,Clagett D,Isaacs C,Elledge R,Horvath A,Hu Y,Li R
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2023
CD11c+ microglia promote white matter repair after ischemic stroke.
Cell death & disease
Jia J,Zheng L,Ye L,Chen J,Shu S,Xu S,Bao X,Xia S,Liu R,Xu Y,Zhang M
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Feb 24 00:00:00 EST 2023
Group 2 Innate Lymphoid Cells Protect Mice from Abdominal Aortic Aneurysm Formation via IL5 and Eosinophils.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Liu T,Deng Z,Fang W,Zhang X,Zhang S,Wang M,Luo S,Meng Z,Liu J,Sukhova GK,Li D,McKenzie ANJ,Libby P,Shi GP,Guo J
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
The ZIP8/SIRT1 axis regulates alveolar progenitor cell renewal in aging and idiopathic pulmonary fibrosis.
The Journal of clinical investigation
Liang J,Huang G,Liu X,Taghavifar F,Liu N,Wang Y,Deng N,Yao C,Xie T,Kulur V,Dai K,Burman A,Rowan SC,Weigt SS,Belperio J,Stripp B,Parks WC,Jiang D,Noble PW
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2022
Interleukin-10 genetically modified clinical-grade mesenchymal stromal cells markedly reinforced functional recovery after spinal cord injury via directing alternative activation of macrophages.
Cellular & molecular biology letters
Gao T,Huang F,Wang W,Xie Y,Wang B
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Mar 17 00:00:00 EDT 2022
BEX1 is a critical determinant of viral myocarditis.
PLoS pathogens
Martens CR,Dorn LE,Kenney AD,Bansal SS,Yount JS,Accornero F
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Feb 01 00:00:00 EST 2022
GPR92 activation in islet macrophages controls β cell function in a diet-induced obesity model.
The Journal of clinical investigation
de Souza CO,Paschoal VA,Sun X,Vishvanath L,Zhang Q,Shao M,Onodera T,Chen S,Joffin N,Bueno LM,Gupta RK,Oh DY
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2022
Mineralocorticoid receptor antagonists and glucocorticoids differentially affect skeletal muscle inflammation and pathology in muscular dystrophy.
JCI insight
Howard ZM,Gomatam CK,Rabolli CP,Lowe J,Piepho AB,Bansal SS,Accornero F,Rafael-Fortney JA
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Oct 10 00:00:00 EDT 2022
Comparative analysis of thoracic and abdominal aortic aneurysms across the segment and species at the single-cell level.
Frontiers in pharmacology
Wu H,Xie C,Wang R,Cheng J,Xu Q,Zhao H
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Feb 02 00:00:00 EST 2023
Microglial angiotensin type 2 receptors mediate sex-specific expression of inflammatory cytokines independently of circulating estrogen.
Glia
Garrido-Gil P,Pedrosa MA,Garcia-Garrote M,Pequeño-Valtierra A,Rodríguez-Castro J,García-Souto D,Rodríguez-Pérez AI,Labandeira-Garcia JL
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2022
Hematopoietic-Mesenchymal Signals Regulate the Properties of Mesenchymal Stem Cells.
International journal of molecular sciences
Kanazawa S,Okada H,Riu D,Mabuchi Y,Akazawa C,Iwata J,Hoshi K,Hikita A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jul 26 00:00:00 EDT 2022
Gene-selective transcription promotes the inhibition of tissue reparative macrophages by TNF.
Life science alliance
Dichtl S,Sanin DE,Koss CK,Willenborg S,Petzold A,Tanzer MC,Dahl A,Kabat AM,Lindenthal L,Zeitler L,Satzinger S,Strasser A,Mann M,Roers A,Eming SA,El Kasmi KC,Pearce EJ,Murray PJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Apr 01 00:00:00 EDT 2022
Intranasal delivery of a small-molecule ErbB inhibitor promotes recovery from acute and late-stage CNS inflammation.
JCI insight
Linnerbauer M,Lößlein L,Vandrey O,Tsaktanis T,Beer A,Naumann UJ,Panier F,Beyer T,Nirschl L,Kuramatsu JB,Winkler J,Quintana FJ,Rothhammer V
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Apr 08 00:00:00 EDT 2022
Tumor collection/processing under physioxia uncovers highly relevant signaling networks and drug sensitivity.
Science advances
Kumar B,Adebayo AK,Prasad M,Capitano ML,Wang R,Bhat-Nakshatri P,Anjanappa M,Simpson E,Chen D,Liu Y,Schilder JM,Colter AB,Maguire C,Temm CJ,Sandusky G,Doud EH,Wijeratne AB,Mosley AL,Broxmeyer HE,Nakshatri H
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Jan 14 00:00:00 EST 2022
The RNA-binding protein IMP2 drives a stromal-Th17 cell circuit in autoimmune neuroinflammation.
JCI insight
Bechara R,Amatya N,Majumder S,Zhou C,Li Y,Liu Q,McGeachy MJ,Gaffen SL
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Feb 08 00:00:00 EST 2022
PI3K signaling specifies proximal-distal fate by driving a developmental gene regulatory network in SOX9+ mouse lung progenitors.
eLife
Khattar D,Fernandes S,Snowball J,Guo M,Gillen MC,Jain SS,Sinner D,Zacharias W,Swarr DT
15-0451-82 was used in Flow cytometry/Cell sorting to analyze paired gene expression (RNA-Seq) and chromatin accessibility (ATAC-Seq) data from SOX9+ epithelial progenitor cells (EPCs) during embryonic development in Mus musculus.
Wed Aug 17 00:00:00 EDT 2022
Induction of alarmin S100A8/A9 mediates activation of aberrant neutrophils in the pathogenesis of COVID-19.
Cell host & microbe
Guo Q,Zhao Y,Li J,Liu J,Yang X,Guo X,Kuang M,Xia H,Zhang Z,Cao L,Luo Y,Bao L,Wang X,Wei X,Deng W,Wang N,Chen L,Chen J,Zhu H,Gao R,Qin C,Wang X,You F
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Feb 10 00:00:00 EST 2021
Stage-specific action of Runx1 and GATA3 controls silencing of PU.1 expression in mouse pro-T cells.
The Journal of experimental medicine
Hosokawa H,Koizumi M,Masuhara K,Romero-Wolf M,Tanaka T,Nakayama T,Rothenberg EV
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Aug 02 00:00:00 EDT 2021
Uptake and trafficking of different sized PLGA nanoparticles by dendritic cells in imiquimod-induced psoriasis-like mice model.
Acta pharmaceutica Sinica. B
Lin Z,Xi L,Chen S,Tao J,Wang Y,Chen X,Li P,Wang Z,Zheng Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
Mapping the origin and fate of myeloid cells in distinct compartments of the eye by single-cell profiling.
The EMBO journal
Wieghofer P,Hagemeyer N,Sankowski R,Schlecht A,Staszewski O,Amann L,Gruber M,Koch J,Hausmann A,Zhang P,Boneva S,Masuda T,Hilgendorf I,Goldmann T,Böttcher C,Priller J,Rossi FM,Lange C,Prinz M
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Mar 15 00:00:00 EDT 2021
Ablation of lysozyme M-positive cells prevents aircraft noise-induced vascular damage without improving cerebral side effects.
Basic research in cardiology
Frenis K,Helmstädter J,Ruan Y,Schramm E,Kalinovic S,Kröller-Schön S,Bayo Jimenez MT,Hahad O,Oelze M,Jiang S,Wenzel P,Sommer CJ,Frauenknecht KBM,Waisman A,Gericke A,Daiber A,Münzel T,Steven S
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Apr 30 00:00:00 EDT 2021
Photoreceptor protection by mesenchymal stem cell transplantation identifies exosomal MiR-21 as a therapeutic for retinal degeneration.
Cell death and differentiation
Deng CL,Hu CB,Ling ST,Zhao N,Bao LH,Zhou F,Xiong YC,Chen T,Sui BD,Yu XR,Hu CH
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Mar 01 00:00:00 EST 2021
Loss of p53 in mesenchymal stem cells promotes alteration of bone remodeling through negative regulation of osteoprotegerin.
Cell death and differentiation
Velletri T,Huang Y,Wang Y,Li Q,Hu M,Xie N,Yang Q,Chen X,Chen Q,Shou P,Gan Y,Candi E,Annicchiarico-Petruzzelli M,Agostini M,Yang H,Melino G,Shi Y,Wang Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
Decreased antiviral immune response within the central nervous system of aged mice is associated with increased lethality of West Nile virus encephalitis.
Aging cell
Funk KE,Arutyunov AD,Desai P,White JP,Soung AL,Rosen SF,Diamond MS,Klein RS
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2021
Anti-adipogenic signals at the onset of obesity-related inflammation in white adipose tissue.
Cellular and molecular life sciences : CMLS
Caputo T,Tran VDT,Bararpour N,Winkler C,Aguileta G,Trang KB,Giordano Attianese GMP,Wilson A,Thomas A,Pagni M,Guex N,Desvergne B,Gilardi F
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
RANKL from bone marrow adipose lineage cells promotes osteoclast formation and bone loss.
EMBO reports
Hu Y,Li X,Zhi X,Cong W,Huang B,Chen H,Wang Y,Li Y,Wang L,Fang C,Guo J,Liu Y,Cui J,Cao L,Weng W,Zhou Q,Wang S,Chen X,Su J
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Jul 05 00:00:00 EDT 2021
Macrophage polarization by MSC-derived CXCL12 determines tumor growth.
Cellular & molecular biology letters
Babazadeh S,Nassiri SM,Siavashi V,Sahlabadi M,Hajinasrollah M,Zamani-Ahmadmahmudi M
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat Jun 26 00:00:00 EDT 2021
The effects of postmortem delay on mouse and human microglia gene expression.
Glia
Heng Y,Dubbelaar ML,Marie SKN,Boddeke EWGM,Eggen BJL
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
Intrinsic DNA damage repair deficiency results in progressive microglia loss and replacement.
Glia
Zhang X,Heng Y,Kooistra SM,van Weering HRJ,Brummer ML,Gerrits E,Wesseling EM,Brouwer N,Nijboer TW,Dubbelaar ML,Boddeke EWGM,Eggen BJL
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Mar 01 00:00:00 EST 2021
Intercellular Interactions of an Adipogenic CXCL12-Expressing Stromal Cell Subset in Murine Bone Marrow.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Matsushita Y,Chu AKY,Ono W,Welch JD,Ono N
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
Macrophage LAMTOR1 Deficiency Prevents Dietary Obesity and Insulin Resistance Through Inflammation-Induced Energy Expenditure.
Frontiers in cell and developmental biology
Ying L,Zhang M,Ma X,Si Y,Li X,Su J,Yin J,Bao Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jun 08 00:00:00 EDT 2021
Alterations of macrophage and neutrophil content in skeletal muscle of aged versus young mice.
Muscle & nerve
Kawanishi N,Machida S
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
Differential activation of Ca2+ influx channels modulate stem cell potency, their proliferation/viability and tissue regeneration.
NPJ Regenerative medicine
Ahamad N,Sun Y,Nascimento Da Conceicao V,Xavier Paul Ezhilan CRD,Natarajan M,Singh BB
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Oct 20 00:00:00 EDT 2021
Suppression of mucosal Th17 memory responses by acellular pertussis vaccines enhances nasal Bordetella pertussis carriage.
NPJ vaccines
Dubois V,Chatagnon J,Thiriard A,Bauderlique-Le Roy H,Debrie AS,Coutte L,Locht C
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Jan 08 00:00:00 EST 2021
Synergistic immunotherapy of glioblastoma by dual targeting of IL-6 and CD40.
Nature communications
Yang F,He Z,Duan H,Zhang D,Li J,Yang H,Dorsey JF,Zou W,Nabavizadeh SA,Bagley SJ,Abdullah K,Brem S,Zhang L,Xu X,Byrne KT,Vonderheide RH,Gong Y,Fan Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jun 08 00:00:00 EDT 2021
The effect of fingolimod on regulatory T cells in a mouse model of brain ischaemia.
Journal of neuroinflammation
Malone K,Diaz Diaz AC,Shearer JA,Moore AC,Waeber C
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat Jan 30 00:00:00 EST 2021
NOX4 regulates macrophage apoptosis resistance to induce fibrotic progression.
The Journal of biological chemistry
Larson-Casey JL,Gu L,Kang J,Dhyani A,Carter AB
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2021
The IRE1/XBP1 signaling axis promotes skeletal muscle regeneration through a cell non-autonomous mechanism.
eLife
Roy A,Tomaz da Silva M,Bhat R,Bohnert KR,Iwawaki T,Kumar A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Nov 23 00:00:00 EST 2021
A decrease in NAD+ contributes to the loss of osteoprogenitors and bone mass with aging.
NPJ aging and mechanisms of disease
Kim HN,Ponte F,Warren A,Ring R,Iyer S,Han L,Almeida M
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
Therapeutic Potential of Mesenchymal Stem Cells in a Pre-Clinical Model of Diabetic Kidney Disease and Obesity.
International journal of molecular sciences
Sávio-Silva C,Soinski-Sousa PE,Simplício-Filho A,Bastos RMC,Beyerstedt S,Rangel ÉB
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Feb 04 00:00:00 EST 2021
Use of a Self-Delivering Anti-CCL3 FANA Oligonucleotide as an Innovative Approach to Target Inflammation after Spinal Cord Injury.
eNeuro
Pelisch N,Rosas Almanza J,Stehlik KE,Aperi BV,Kroner A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Jun 18 00:00:00 EDT 2021
T cell-depleting nanoparticles ameliorate bone loss by reducing activated T cells and regulating the Treg/Th17 balance.
Bioactive materials
Yang X,Zhou F,Yuan P,Dou G,Liu X,Liu S,Wang X,Jin R,Dong Y,Zhou J,Lv Y,Deng Z,Liu S,Chen X,Han Y,Jin Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2021
Placenta-Expanded Stromal Cell Therapy in a Rodent Model of Simulated Weightlessness.
Cells
Rubinstein L,Paul AM,Houseman C,Abegaz M,Tabares Ruiz S,O'Neil N,Kunis G,Ofir R,Cohen J,Ronca AE,Globus RK,Tahimic CGT
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Apr 19 00:00:00 EDT 2021
Apoptotic vesicles restore liver macrophage homeostasis to counteract type 2 diabetes.
Journal of extracellular vesicles
Zheng C,Sui B,Zhang X,Hu J,Chen J,Liu J,Wu D,Ye Q,Xiang L,Qiu X,Liu S,Deng Z,Zhou J,Liu S,Shi S,Jin Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat May 01 00:00:00 EDT 2021
Distinct skeletal stem cell types orchestrate long bone skeletogenesis.
eLife
Ambrosi TH,Sinha R,Steininger HM,Hoover MY,Murphy MP,Koepke LS,Wang Y,Lu WJ,Morri M,Neff NF,Weissman IL,Longaker MT,Chan CK
15-0451-82 was used in Flow Cytometry to address previously unappreciated shortcomings of SSC research.
Mon Jul 19 00:00:00 EDT 2021
Distinct skeletal stem cell types orchestrate long bone skeletogenesis.
eLife
Ambrosi TH,Sinha R,Steininger HM,Hoover MY,Murphy MP,Koepke LS,Wang Y,Lu WJ,Morri M,Neff NF,Weissman IL,Longaker MT,Chan CK
15-0451-82 was used in Flow Cytometry to address previously unappreciated shortcomings of SSC research.
Mon Jul 19 00:00:00 EDT 2021
Integrated OMICs unveil the bone-marrow microenvironment in human leukemia.
Cell reports
Passaro D,Garcia-Albornoz M,Diana G,Chakravarty P,Ariza-McNaughton L,Batsivari A,Borràs-Eroles C,Abarrategi A,Waclawiczek A,Ombrato L,Malanchi I,Gribben J,Bonnet D
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue May 11 00:00:00 EDT 2021
PTENα functions as an immune suppressor and promotes immune resistance in PTEN-mutant cancer.
Nature communications
Sun Y,Lu D,Yin Y,Song J,Liu Y,Hao W,Qi F,Zhang G,Zhang X,Liu L,Lin Z,Liang H,Zhao X,Jin Y,Yin Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Aug 26 00:00:00 EDT 2021
Cardiac-derived TGF-β1 confers resistance to diet-induced obesity through the regulation of adipocyte size and function.
Molecular metabolism
Longenecker JZ,Petrosino JM,Martens CR,Hinger SA,Royer CJ,Dorn LE,Branch DA,Serrano J,Stanford KI,Kyriazis GA,Baskin KK,Accornero F
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Dec 01 00:00:00 EST 2021
Determining the effect of aging, recovery time, and post-stroke memantine treatment on delayed thalamic gliosis after cortical infarct.
Scientific reports
Kim GS,Stephenson JM,Al Mamun A,Wu T,Goss MG,Min JW,Li J,Liu F,Marrelli SP
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jun 15 00:00:00 EDT 2021
Cytochrome P450 26A1 Modulates the Polarization of Uterine Macrophages During the Peri-Implantation Period.
Frontiers in immunology
Ji WH,Li DD,Wei DP,Gu AQ,Yang Y,Peng JP
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Feb 10 00:00:00 EST 2022
A molybdenum oxide-based degradable nanosheet for combined chemo-photothermal therapy to improve tumor immunosuppression and suppress distant tumors and lung metastases.
Journal of nanobiotechnology
Qiu N,Yang X,Zhang Y,Zhang J,Ji J,Zhang Y,Kong X,Xi Y,Liu D,Ye L,Zhai G
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sun Dec 19 00:00:00 EST 2021
Visceral adipose tissue imparts peripheral macrophage influx into the hypothalamus.
Journal of neuroinflammation
Chen KE,Lainez NM,Nair MG,Coss D
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Jun 21 00:00:00 EDT 2021
Quantitative in vivo assessment of amyloid-beta phagocytic capacity in an Alzheimer's disease mouse model.
STAR protocols
Lau SF,Wu W,Seo H,Fu AKY,Ip NY
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Mar 19 00:00:00 EDT 2021
Mice with induced pulmonary morbidities display severe lung inflammation and mortality following exposure to SARS-CoV-2.
JCI insight
Falach R,Bar-On L,Lazar S,Kadar T,Mazor O,Aftalion M,Gur D,Evgy Y,Shifman O,Aminov T,Israeli O,Cohen-Gihon I,Zaide G,Gutman H,Vagima Y,Makdasi E,Stein D,Rosenfeld R,Alcalay R,Zahavy E,Levy H,Glinert I,Ben-Shmuel A,Israely T,Melamed S,Politi B,Achdout H,Yitzhaki S,Kronman C,Sabo T
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jun 22 00:00:00 EDT 2021
Epinephrine Production in Th17 Cells and Experimental Autoimmune Encephalitis.
Frontiers in immunology
Yang P,Tian H,Zou YR,Chambon P,Ichinose H,Honig G,Diamond B,Kim SJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Jul 05 00:00:00 EDT 2021
Systemic administration of β-glucan induces immune training in microglia.
Journal of neuroinflammation
Heng Y,Zhang X,Borggrewe M,van Weering HRJ,Brummer ML,Nijboer TW,Joosten LAB,Netea MG,Boddeke EWGM,Laman JD,Eggen BJL
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Feb 22 00:00:00 EST 2021
Macrophages Maintain Epithelium Integrity by Limiting Fungal Product Absorption.
Cell
Chikina AS,Nadalin F,Maurin M,San-Roman M,Thomas-Bonafos T,Li XV,Lameiras S,Baulande S,Henri S,Malissen B,Lacerda Mariano L,Barbazan J,Blander JM,Iliev ID,Matic Vignjevic D,Lennon-Duménil AM
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Oct 15 00:00:00 EDT 2020
The Cytokine TGF-β Induces Interleukin-31 Expression from Dermal Dendritic Cells to Activate Sensory Neurons and Stimulate Wound Itching.
Immunity
Xu J,Zanvit P,Hu L,Tseng PY,Liu N,Wang F,Liu O,Zhang D,Jin W,Guo N,Han Y,Yin J,Cain A,Hoon MA,Wang S,Chen W
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Aug 18 00:00:00 EDT 2020
Exposure to diesel exhaust particles increases susceptibility to invasive pneumococcal disease.
The Journal of allergy and clinical immunology
Shears RK,Jacques LC,Naylor G,Miyashita L,Khandaker S,Lebre F,Lavelle EC,Grigg J,French N,Neill DR,Kadioglu A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2020
Doxycycline Reduces Scar Thickness and Improves Collagen Architecture.
Annals of surgery
Moore AL,desJardins-Park HE,Duoto BA,Mascharak S,Murphy MP,Irizarry DM,Foster DS,Jones RE,Barnes LA,Marshall CD,Ransom RC,Wernig G,Longaker MT
15-0451 was used in Flow cytometry/Cell sorting to investigate the effects of local doxycycline administration on skin scarring.
Wed Jul 01 00:00:00 EDT 2020
Modulation of the functions of myeloid-derived suppressor cells : a new strategy of hydrogen sulfide anti-cancer effects.
British journal of pharmacology
De Cicco P,Ercolano G,Rubino V,Terrazzano G,Ruggiero G,Cirino G,Ianaro A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat Feb 01 00:00:00 EST 2020
Membrane expression of the estrogen receptor ERα is required for intercellular communications in the mammary epithelium.
Development (Cambridge, England)
Gagniac L,Rusidzé M,Boudou F,Cagnet S,Adlanmerini M,Jeannot P,Gaide N,Giton F,Besson A,Weyl A,Gourdy P,Raymond-Letron I,Arnal JF,Brisken C,Lenfant F
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Mar 11 00:00:00 EDT 2020
Dysfunction of B Cell Leading to Failure of Immunoglobulin Response Is Ameliorated by Dietary Silk Peptide in 14-Month-Old C57BL/6 Mice.
Frontiers in nutrition
Chei S,Oh HJ,Lee K,Jin H,Lee JY,Lee BY
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Dec 18 00:00:00 EST 2020
Sema3A and HIF1α co-overexpressed iPSC-MSCs/HA scaffold facilitates the repair of calvarial defect in a mouse model.
Journal of cellular physiology
Li J,Wang T,Li C,Wang Z,Wang P,Zheng L
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2020
Detection of Synaptic Proteins in Microglia by Flow Cytometry.
Frontiers in molecular neuroscience
Brioschi S,d'Errico P,Amann LS,Janova H,Wojcik SM,Meyer-Luehmann M,Rajendran L,Wieghofer P,Paolicelli RC,Biber K
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Nov 03 00:00:00 EST 2020
Extracellular HMGB-1 activates inflammatory signaling in tendon cells and tissues.
Therapeutic advances in chronic disease
Zhang C,Gu X,Zhao G,Wang W,Shao J,Zhu J,Yuan T,Sun J,Nie D,Zhou Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sun Apr 17 00:00:00 EDT 2022
Resveratrol rescues TNF‑α‑induced inhibition of osteogenesis in human periodontal ligament stem cells via the ERK1/2 pathway.
Molecular medicine reports
Yuan J,Wang X,Ma D,Gao H,Zheng D,Zhang J
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri May 01 00:00:00 EDT 2020
Enhancer remodeling promotes tumor-initiating activity in NRF2-activated non-small cell lung cancers.
Nature communications
Okazaki K,Anzawa H,Liu Z,Ota N,Kitamura H,Onodera Y,Alam MM,Matsumaru D,Suzuki T,Katsuoka F,Tadaka S,Motoike I,Watanabe M,Hayasaka K,Sakurada A,Okada Y,Yamamoto M,Suzuki T,Kinoshita K,Sekine H,Motohashi H
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Nov 20 00:00:00 EST 2020
Eosinophils improve cardiac function after myocardial infarction.
Nature communications
Liu J,Yang C,Liu T,Deng Z,Fang W,Zhang X,Li J,Huang Q,Liu C,Wang Y,Yang D,Sukhova GK,Lindholt JS,Diederichsen A,Rasmussen LM,Li D,Newton G,Luscinskas FW,Liu L,Libby P,Wang J,Guo J,Shi GP
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Dec 16 00:00:00 EST 2020
Ly6cLo non-classical monocytes promote resolution of rhesus rotavirus-mediated perinatal hepatic inflammation.
Scientific reports
Alkhani A,Levy CS,Tsui M,Rosenberg KA,Polovina K,Mattis AN,Mack M,Van Dyken S,Wang BM,Maher JJ,Nijagal A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Apr 28 00:00:00 EDT 2020
Stromal CCL2 Signaling Promotes Mammary Tumor Fibrosis through Recruitment of Myeloid-Lineage Cells.
Cancers
Kuziel G,Thompson V,D'Amato JV,Arendt LM
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jul 28 00:00:00 EDT 2020
Small GTPase RAB6 deficiency promotes alveolar progenitor cell renewal and attenuates PM2.5-induced lung injury and fibrosis.
Cell death & disease
Yang L,Liu G,Li X,Xia Z,Wang Y,Lin W,Zhang W,Zhang W,Li X
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sun Oct 04 00:00:00 EDT 2020
A RUNX2 stabilization pathway mediates physiologic and pathologic bone formation.
Nature communications
Kim JM,Yang YS,Park KH,Ge X,Xu R,Li N,Song M,Chun H,Bok S,Charles JF,Filhol-Cochet O,Boldyreff B,Dinter T,Yu PB,Kon N,Gu W,Takarada T,Greenblatt MB,Shim JH
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri May 08 00:00:00 EDT 2020
Analysis of the immune response to sciatic nerve injury identifies efferocytosis as a key mechanism of nerve debridement.
eLife
Kalinski AL,Yoon C,Huffman LD,Duncker PC,Kohen R,Passino R,Hafner H,Johnson C,Kawaguchi R,Carbajal KS,Jara JS,Hollis E,Geschwind DH,Segal BM,Giger RJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Dec 02 00:00:00 EST 2020
Resolvin D1 supports skeletal myofiber regeneration via actions on myeloid and muscle stem cells.
JCI insight
Markworth JF,Brown LA,Lim E,Floyd C,Larouche J,Castor-Macias JA,Sugg KB,Sarver DC,Macpherson PC,Davis C,Aguilar CA,Maddipati KR,Brooks SV
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2020
Chronic alcohol-induced neuroinflammation involves CCR2/5-dependent peripheral macrophage infiltration and microglia alterations.
Journal of neuroinflammation
Lowe PP,Morel C,Ambade A,Iracheta-Vellve A,Kwiatkowski E,Satishchandran A,Furi I,Cho Y,Gyongyosi B,Catalano D,Lefebvre E,Fischer L,Seyedkazemi S,Schafer DP,Szabo G
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Oct 09 00:00:00 EDT 2020
Inhibition of formyl peptide receptors improves the outcome in a mouse model of Alzheimer disease.
Journal of neuroinflammation
Schröder N,Schaffrath A,Welter JA,Putzka T,Griep A,Ziegler P,Brandt E,Samer S,Heneka MT,Kaddatz H,Zhan J,Kipp E,Pufe T,Tauber SC,Kipp M,Brandenburg LO
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Apr 24 00:00:00 EDT 2020
Ovarian granulosa cell tumor characterization identifies FOXL2 as an immunotherapeutic target.
JCI insight
Pierini S,Tanyi JL,Simpkins F,George E,Uribe-Herranz M,Drapkin R,Burger R,Morgan MA,Facciabene A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Aug 20 00:00:00 EDT 2020
Butyrophilin-like proteins display combinatorial diversity in selecting and maintaining signature intraepithelial γδ T cell compartments.
Nature communications
Jandke A,Melandri D,Monin L,Ushakov DS,Laing AG,Vantourout P,East P,Nitta T,Narita T,Takayanagi H,Feederle R,Hayday A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jul 28 00:00:00 EDT 2020
Tankyrase inhibition sensitizes melanoma to PD-1 immune checkpoint blockade in syngeneic mouse models.
Communications biology
Waaler J,Mygland L,Tveita A,Strand MF,Solberg NT,Olsen PA,Aizenshtadt A,Fauskanger M,Lund K,Brinch SA,Lycke M,Dybing E,Nygaard V,Bøe SL,Heintz KM,Hovig E,Hammarström C,Corthay A,Krauss S
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Apr 24 00:00:00 EDT 2020
PPARγ inhibition boosts efficacy of PD-L1 Checkpoint Blockade Immunotherapy against Murine Melanoma in a sexually dimorphic manner.
International journal of biological sciences
Wu B,Sun X,Yuan B,Ge F,Gupta HB,Chiang HC,Li J,Hu Y,Curiel TJ,Li R
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Aug 23 00:00:00 EDT 2021
A Synthetic Small Molecule F240B Decreases NLRP3 Inflammasome Activation by Autophagy Induction.
Frontiers in immunology
Wu CH,Gan CH,Li LH,Chang JC,Chen ST,Menon MP,Cheng SM,Yang SP,Ho CL,Chernikov OV,Lin CH,Lam Y,Hua KF
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Jun 23 00:00:00 EDT 2021
CNS Immune Profiling in a Dengue Virus-Infected Immunocompetent Outbred ICR Mice Strain.
Frontiers in cellular and infection microbiology
Shen TJ,Chen CL,Jhan MK,Tseng PC,Lin CF
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Jun 17 00:00:00 EDT 2021
Apoptotic bodies derived from mesenchymal stem cells promote cutaneous wound healing via regulating the functions of macrophages.
Stem cell research & therapy
Liu J,Qiu X,Lv Y,Zheng C,Dong Y,Dou G,Zhu B,Liu A,Wang W,Zhou J,Liu S,Liu S,Gao B,Jin Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Nov 27 00:00:00 EST 2020
Phosphatase SHP1 impedes mesenchymal stromal cell immunosuppressive capacity modulated by JAK1/STAT3 and P38 signals.
Cell & bioscience
Jiang M,Ye J,Wang X,Li N,Wang Y,Shi Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Apr 14 00:00:00 EDT 2022
Mesenchymal Stromal Cell-Derived Exosomes Contribute to Epithelial Regeneration in Experimental Inflammatory Bowel Disease.
Cellular and molecular gastroenterology and hepatology
Barnhoorn MC,Plug L,Jonge ESMM,Molenkamp D,Bos E,Schoonderwoerd MJA,Corver WE,van der Meulen-de Jong AE,Verspaget HW,Hawinkels LJAC
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon May 10 00:00:00 EDT 2021
The ROP16III-dependent early immune response determines the subacute CNS immune response and type III Toxoplasma gondii survival.
PLoS pathogens
Tuladhar S,Kochanowsky JA,Bhaskara A,Ghotmi Y,Chandrasekaran S,Koshy AA
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2019
Distinct Tie2 tyrosine phosphorylation sites dictate phenotypic switching in endothelial progenitor cells.
Journal of cellular physiology
Siavashi V,Nassiri SM,Rahbarghazi R,Mohseni Z,Sharifi AM
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed May 01 00:00:00 EDT 2019
Protumoral bone marrow-derived cells migrate via Gβγ-dependent signaling pathways and exhibit a complex repertoire of RhoGEFs.
Journal of cell communication and signaling
Cervantes-Villagrana RD,Color-Aparicio VM,Reyes-Cruz G,Vázquez-Prado J
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat Jun 01 00:00:00 EDT 2019
Antiallodynic Effects of Cannabinoid Receptor 2 (CB2R) Agonists on Retrovirus Infection-Induced Neuropathic Pain.
Pain research & management
Sheng WS,Chauhan P,Hu S,Prasad S,Lokensgard JR
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Dec 16 00:00:00 EST 2019
Antitumor and immunomodulatory effects of a novel multitarget inhibitor, CS2164, in mouse hepatocellular carcinoma models.
Anti-cancer drugs
Zhou Y,Fu C,Kong Y,Pan D,Wang Y,Huang S,Li Z,Ning Z,Lu X,Shan S,Xin L
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2019
Atherogenesis in the Carotid Artery with and without Interrupted Blood Flow of Two Hyperlipidemic Mouse Strains.
Journal of vascular research
Zhao J,Huangfu C,Chang Z,Grainger AT,Liu Z,Shi W
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Jan 27 00:00:00 EST 2020
Exposure to Cigarette Smoke Augments Post-ischemic Brain Injury and Inflammation via Mobilization of Neutrophils and Monocytes.
Frontiers in immunology
Li H,Li X,Gao S,Wang D,Gao X,Li Y,Wang X,Cui Z,Ma H,Liu Q,Li M
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
CSF1R antagonism limits local restimulation of antiviral CD8+ T cells during viral encephalitis.
Journal of neuroinflammation
Funk KE,Klein RS
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Jan 31 00:00:00 EST 2019
Autophagy regulates inflammatory programmed cell death via turnover of RHIM-domain proteins.
eLife
Lim J,Park H,Heisler J,Maculins T,Roose-Girma M,Xu M,Mckenzie B,van Lookeren Campagne M,Newton K,Murthy A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jul 09 00:00:00 EDT 2019
The microRNAs miR-204 and miR-211 maintain joint homeostasis and protect against osteoarthritis progression.
Nature communications
Huang J,Zhao L,Fan Y,Liao L,Ma PX,Xiao G,Chen D
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Jun 28 00:00:00 EDT 2019
MDM2 inhibitor APG-115 synergizes with PD-1 blockade through enhancing antitumor immunity in the tumor microenvironment.
Journal for immunotherapy of cancer
Fang DD,Tang Q,Kong Y,Wang Q,Gu J,Fang X,Zou P,Rong T,Wang J,Yang D,Zhai Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Nov 28 00:00:00 EST 2019
Muscularis Propria Macrophages Alter the Proportion of Nitrergic but Not Cholinergic Gastric Myenteric Neurons.
Cellular and molecular gastroenterology and hepatology
Cipriani G,Terhaar ML,Eisenman ST,Ji S,Linden DR,Wright AM,Sha L,Ordog T,Szurszewski JH,Gibbons SJ,Farrugia G
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jun 04 00:00:00 EDT 2019
Neutrophils promote CXCR3-dependent itch in the development of atopic dermatitis.
eLife
Walsh CM,Hill RZ,Schwendinger-Schreck J,Deguine J,Brock EC,Kucirek N,Rifi Z,Wei J,Gronert K,Brem RB,Barton GM,Bautista DM
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Oct 21 00:00:00 EDT 2019
NLRP3 inflammasome in fibroblasts links tissue damage with inflammation in breast cancer progression and metastasis.
Nature communications
Ershaid N,Sharon Y,Doron H,Raz Y,Shani O,Cohen N,Monteran L,Leider-Trejo L,Ben-Shmuel A,Yassin M,Gerlic M,Ben-Baruch A,Pasmanik-Chor M,Apte R,Erez N
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Sep 26 00:00:00 EDT 2019
Dietary calories and lipids synergistically shape adipose tissue cellularity during postnatal growth.
Molecular metabolism
Meln I,Wolff G,Gajek T,Koddebusch J,Lerch S,Harbrecht L,Hong W,Bayindir-Buchhalter I,Krunic D,Augustin HG,Vegiopoulos A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat Jun 01 00:00:00 EDT 2019
FAK activity sustains intrinsic and acquired ovarian cancer resistance to platinum chemotherapy.
eLife
Diaz Osterman CJ,Ozmadenci D,Kleinschmidt EG,Taylor KN,Barrie AM,Jiang S,Bean LM,Sulzmaier FJ,Jean C,Tancioni I,Anderson K,Uryu S,Cordasco EA,Li J,Chen XL,Fu G,Ojalill M,Rappu P,Heino J,Mark AM,Xu G,Fisch KM,Kolev VN,Weaver DT,Pachter JA,Győrffy B,McHale MT,Connolly DC,Molinolo A,Stupack DG,Schlaepfer DD
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Sep 03 00:00:00 EDT 2019
PDGFRα+ stromal adipocyte progenitors transition into epithelial cells during lobulo-alveologenesis in the murine mammary gland.
Nature communications
Joshi PA,Waterhouse PD,Kasaian K,Fang H,Gulyaeva O,Sul HS,Boutros PC,Khokha R
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Apr 15 00:00:00 EDT 2019
Growth Factor Screening in Dystrophic Muscles Reveals PDGFB/PDGFRB-Mediated Migration of Interstitial Stem Cells.
International journal of molecular sciences
Camps J,Grosemans H,Gijsbers R,Maes C,Sampaolesi M
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Mar 05 00:00:00 EST 2019
Mitophagy in Intestinal Epithelial Cells Triggers Adaptive Immunity during Tumorigenesis.
Cell
Ziegler PK,Bollrath J,Pallangyo CK,Matsutani T,Canli Ö,De Oliveira T,Diamanti MA,Müller N,Gamrekelashvili J,Putoczki T,Horst D,Mankan AK,Öner MG,Müller S,Müller-Höcker J,Kirchner T,Slotta-Huspenina J,Taketo MM,Reinheckel T,Dröse S,Larner AC,Wels WS,Ernst M,Greten TF,Arkan MC,Korn T,Wirth D,Greten FR
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Jun 28 00:00:00 EDT 2018
Chanzyme TRPM7 Mediates the Ca2+ Influx Essential for Lipopolysaccharide-Induced Toll-Like Receptor 4 Endocytosis and Macrophage Activation.
Immunity
Schappe MS,Szteyn K,Stremska ME,Mendu SK,Downs TK,Seegren PV,Mahoney MA,Dixit S,Krupa JK,Stipes EJ,Rogers JS,Adamson SE,Leitinger N,Desai BN
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jan 16 00:00:00 EST 2018
E3 Ligase VHL Promotes Group 2 Innate Lymphoid Cell Maturation and Function via Glycolysis Inhibition and Induction of Interleukin-33 Receptor.
Immunity
Li Q,Li D,Zhang X,Wan Q,Zhang W,Zheng M,Zou L,Elly C,Lee JH,Liu YC
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Feb 20 00:00:00 EST 2018
Muscle Satellite Cell Cross-Talk with a Vascular Niche Maintains Quiescence via VEGF and Notch Signaling.
Cell stem cell
Verma M,Asakura Y,Murakonda BSR,Pengo T,Latroche C,Chazaud B,McLoon LK,Asakura A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Oct 04 00:00:00 EDT 2018
Endothelial cells in the innate response to allergens and initiation of atopic asthma.
The Journal of clinical investigation
Asosingh K,Weiss K,Queisser K,Wanner N,Yin M,Aronica M,Erzurum S
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Jul 02 00:00:00 EDT 2018
Defective germline reprogramming rewires the spermatogonial transcriptome.
Nature structural & molecular biology
Vasiliauskaitė L,Berrens RV,Ivanova I,Carrieri C,Reik W,Enright AJ,O'Carroll D
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue May 01 00:00:00 EDT 2018
11 Beta-hydroxysteroid dehydrogenase type 1 regulates synovitis, joint destruction, and systemic bone loss in chronic polyarthritis.
Journal of autoimmunity
Hardy RS,Fenton C,Croft AP,Naylor AJ,Begum R,Desanti G,Buckley CD,Lavery G,Cooper MS,Raza K
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2018
Inhibition of the JNK/MAPK signaling pathway by myogenesis-associated miRNAs is required for skeletal muscle development.
Cell death and differentiation
Xie SJ,Li JH,Chen HF,Tan YY,Liu SR,Zhang Y,Xu H,Yang JH,Liu S,Zheng LL,Huang MB,Guo YH,Zhang Q,Zhou H,Qu LH
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2018
VISTA expression by microglia decreases during inflammation and is differentially regulated in CNS diseases.
Glia
Borggrewe M,Grit C,Den Dunnen WFA,Burm SM,Bajramovic JJ,Noelle RJ,Eggen BJL,Laman JD
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
Protease-activated receptor-2 signaling through β-arrestin-2 mediates Alternaria alkaline serine protease-induced airway inflammation.
American journal of physiology. Lung cellular and molecular physiology
Yee MC,Nichols HL,Polley D,Saifeddine M,Pal K,Lee K,Wilson EH,Daines MO,Hollenberg MD,Boitano S,DeFea KA
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
MyD88 is required for satellite cell-mediated myofiber regeneration in dystrophin-deficient mdx mice.
Human molecular genetics
Gallot YS,Straughn AR,Bohnert KR,Xiong G,Hindi SM,Kumar A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2018
mTOR Modulates CD8+ T Cell Differentiation in Mice with Invasive Pulmonary Aspergillosis.
Open life sciences
Wang H,Xiao Y,Su L,Cui N,Liu D
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
MT4-MMP deficiency increases patrolling monocyte recruitment to early lesions and accelerates atherosclerosis.
Nature communications
Clemente C,Rius C,Alonso-Herranz L,Martín-Alonso M,Pollán Á,Camafeita E,Martínez F,Mota RA,Núñez V,Rodríguez C,Seiki M,Martínez-González J,Andrés V,Ricote M,Arroyo AG
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Mar 02 00:00:00 EST 2018
Lsd1 regulates skeletal muscle regeneration and directs the fate of satellite cells.
Nature communications
Tosic M,Allen A,Willmann D,Lepper C,Kim J,Duteil D,Schüle R
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Jan 25 00:00:00 EST 2018
Diet-Induced Obesity Elicits Macrophage Infiltration and Reduction in Spine Density in the Hypothalami of Male but Not Female Mice.
Frontiers in immunology
Lainez NM,Jonak CR,Nair MG,Ethell IM,Wilson EH,Carson MJ,Coss D
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Sep 25 00:00:00 EDT 2019
Adipocyte hypertrophy and lipid dynamics underlie mammary gland remodeling after lactation.
Nature communications
Zwick RK,Rudolph MC,Shook BA,Holtrup B,Roth E,Lei V,Van Keymeulen A,Seewaldt V,Kwei S,Wysolmerski J,Rodeheffer MS,Horsley V
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Sep 04 00:00:00 EDT 2018
ATF3 Sustains IL-22-Induced STAT3 Phosphorylation to Maintain Mucosal Immunity Through Inhibiting Phosphatases.
Frontiers in immunology
Glal D,Sudhakar JN,Lu HH,Liu MC,Chiang HY,Liu YC,Cheng CF,Shui JW
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Sep 26 00:00:00 EDT 2019
Stem cell factor is selectively secreted by arterial endothelial cells in bone marrow.
Nature communications
Xu C,Gao X,Wei Q,Nakahara F,Zimmerman SE,Mar J,Frenette PS
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Jun 22 00:00:00 EDT 2018
Rap1 deficiency-provoked paracrine dysfunction impairs immunosuppressive potency of mesenchymal stem cells in allograft rejection of heart transplantation.
Cell death & disease
Ding Y,Liang X,Zhang Y,Yi L,Shum HC,Chen Q,Chan BP,Fan H,Liu Z,Tergaonkar V,Qi Z,Tse HF,Lian Q
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Mar 07 00:00:00 EST 2018
Partial depletion of CD206-positive M2-like macrophages induces proliferation of beige progenitors and enhances browning after cold stimulation.
Scientific reports
Igarashi Y,Nawaz A,Kado T,Bilal M,Kuwano T,Yamamoto S,Sasahara M,Jiuxiang X,Inujima A,Koizumi K,Imura J,Shibahara N,Usui I,Fujisaka S,Tobe K
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2018
Differential cytokine contributions of perivascular haematopoietic stem cell niches.
Nature cell biology
Asada N,Kunisaki Y,Pierce H,Wang Z,Fernandez NF,Birbrair A,Ma'ayan A,Frenette PS
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2017
A CCR4 antagonist reverses the tumor-promoting microenvironment of renal cancer.
The Journal of clinical investigation
Berlato C,Khan MN,Schioppa T,Thompson R,Maniati E,Montfort A,Jangani M,Canosa M,Kulbe H,Hagemann UB,Duncan AR,Fletcher L,Wilkinson RW,Powles T,Quezada SA,Balkwill FR
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2017
Anti-CD137 Suppresses Tumor Growth by Blocking Reverse Signaling by CD137 Ligand.
Cancer research
Kang SW,Lee SC,Park SH,Kim J,Kim HH,Lee HW,Seo SK,Kwon BS,Cho HR,Kwon B
15-0451-82 was used in Flow Cytometry to report findings related to reverse signaling by CD137 ligand (CD137L) in antigen-presenting dendritic cells (DC) in tumors that address these paradoxical results.
Wed Nov 01 00:00:00 EDT 2017
Oncolytic Adenovirus and Tumor-Targeting Immune Modulatory Therapy Improve Autologous Cancer Vaccination.
Cancer research
Jiang H,Rivera-Molina Y,Gomez-Manzano C,Clise-Dwyer K,Bover L,Vence LM,Yuan Y,Lang FF,Toniatti C,Hossain MB,Fueyo J
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat Jul 15 00:00:00 EDT 2017
Sigma-1 receptors control immune-driven peripheral opioid analgesia during inflammation in mice.
Proceedings of the National Academy of Sciences of the United States of America
Tejada MA,Montilla-García A,Cronin SJ,Cikes D,Sánchez-Fernández C,González-Cano R,Ruiz-Cantero MC,Penninger JM,Vela JM,Baeyens JM,Cobos EJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
Obesity reversibly depletes the basal cell population and enhances mammary epithelial cell estrogen receptor alpha expression and progenitor activity.
Breast cancer research : BCR
Chamberlin T,D'Amato JV,Arendt LM
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Nov 29 00:00:00 EST 2017
Depletion of microglia exacerbates postischemic inflammation and brain injury.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Jin WN,Shi SX,Li Z,Li M,Wood K,Gonzales RJ,Liu Q
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
An atypical role for the myeloid receptor Mincle in central nervous system injury.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Arumugam TV,Manzanero S,Furtado M,Biggins PJ,Hsieh YH,Gelderblom M,MacDonald KP,Salimova E,Li YI,Korn O,Dewar D,Macrae IM,Ashman RB,Tang SC,Rosenthal NA,Ruitenberg MJ,Magnus T,Wells CA
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Role of Triggering Receptor Expressed on Myeloid Cell-1 Expression in Mammalian Target of Rapamycin Modulation of CD8+ T-cell Differentiation during the Immune Response to Invasive Pulmonary Aspergillosis.
Chinese medical journal
Cui N,Wang H,Su LX,Zhang JH,Long Y,Liu DW
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat May 20 00:00:00 EDT 2017
Cornea lymphatics drive the CD8+ T-cell response to herpes simplex virus-1.
Immunology and cell biology
Gurung HR,Carr MM,Carr DJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sun Jan 01 00:00:00 EST 2017
Cytochrome P450 26A1 modulates natural killer cells in mouse early pregnancy.
Journal of cellular and molecular medicine
Meng CY,Li ZY,Fang WN,Song ZH,Yang DD,Li DD,Yang Y,Peng JP
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
β3-Adrenergic Regulation of EPC Features Through Manipulation of the Bone Marrow MSC Niche.
Journal of cellular biochemistry
Vafaei R,Nassiri SM,Siavashi V
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2017
The common parasite Toxoplasma gondii induces prostatic inflammation and microglandular hyperplasia in a mouse model.
The Prostate
Colinot DL,Garbuz T,Bosland MC,Wang L,Rice SE,Sullivan WJ,Arrizabalaga G,Jerde TJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat Jul 01 00:00:00 EDT 2017
The PERK arm of the unfolded protein response regulates satellite cell-mediated skeletal muscle regeneration.
eLife
Xiong G,Hindi SM,Mann AK,Gallot YS,Bohnert KR,Cavener DR,Whittemore SR,Kumar A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Mar 23 00:00:00 EDT 2017
microRNA-449a modulates medullary thymic epithelial cell differentiation.
Scientific reports
Chen P,Zhang H,Sun X,Hu Y,Jiang W,Liu Z,Liu S,Zhang X
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Nov 21 00:00:00 EST 2017
Tristetraprolin expression by keratinocytes controls local and systemic inflammation.
JCI insight
Andrianne M,Assabban A,La C,Mogilenko D,Salle DS,Fleury S,Doumont G,Van Simaeys G,Nedospasov SA,Blackshear PJ,Dombrowicz D,Goriely S,Van Maele L
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Jun 02 00:00:00 EDT 2017
The Interleukin (IL)-1R1 pathway is a critical negative regulator of PyMT-mediated mammary tumorigenesis and pulmonary metastasis.
Oncoimmunology
Dagenais M,Dupaul-Chicoine J,Douglas T,Champagne C,Morizot A,Saleh M
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Zoledronate suppressed angiogenesis and osteogenesis by inhibiting osteoclasts formation and secretion of PDGF-BB.
PloS one
Gao SY,Zheng GS,Wang L,Liang YJ,Zhang SE,Lao XM,Li K,Liao GQ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Sep 18 00:00:00 EDT 2017
Irgm1 coordinately regulates autoimmunity and host defense at select mucosal surfaces.
JCI insight
Azzam KM,Madenspacher JH,Cain DW,Lai L,Gowdy KM,Rai P,Janardhan K,Clayton N,Cunningham W,Jensen H,Patel PS,Kearney JF,Taylor GA,Fessler MB
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Aug 17 00:00:00 EDT 2017
Cardiosphere-Derived Cells Require Endoglin for Paracrine-Mediated Angiogenesis.
Stem cell reports
Redgrave RE,Tual-Chalot S,Davison BJ,Singh E,Hall D,Amirrasouli MM,Gilchrist D,Medvinsky A,Arthur HM
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue May 09 00:00:00 EDT 2017
Classical dendritic cells are required for dietary antigen-mediated induction of peripheral T(reg) cells and tolerance.
Nature immunology
Esterházy D,Loschko J,London M,Jove V,Oliveira TY,Mucida D
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sun May 01 00:00:00 EDT 2016
Oligodendrocyte death results in immune-mediated CNS demyelination.
Nature neuroscience
Traka M,Podojil JR,McCarthy DP,Miller SD,Popko B
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2016
The breast tumor microenvironment alters the phenotype and function of natural killer cells.
Cellular & molecular immunology
Krneta T,Gillgrass A,Chew M,Ashkar AA
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Sep 01 00:00:00 EDT 2016
Non-Cell-Autonomous Regulation of Prostate Epithelial Homeostasis by Androgen Receptor.
Molecular cell
Zhang B,Kwon OJ,Henry G,Malewska A,Wei X,Zhang L,Brinkley W,Zhang Y,Castro PD,Titus M,Chen R,Sayeeduddin M,Raj GV,Mauck R,Roehrborn C,Creighton CJ,Strand DW,Ittmann MM,Xin L
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Sep 15 00:00:00 EDT 2016
Lymphatic Contribution to the Cellular Niche in Heterotopic Ossification.
Annals of surgery
Loder S,Agarwal S,Sorkin M,Breuler C,Li J,Peterson J,Gardenier J,Hsieh HH,Wang SC,Mehrara BJ,Levi B
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
Analysis of Bone-Cartilage-Stromal Progenitor Populations in Trauma Induced and Genetic Models of Heterotopic Ossification.
Stem cells (Dayton, Ohio)
Agarwal S,Loder SJ,Sorkin M,Li S,Shrestha S,Zhao B,Mishina Y,James AW,Levi B
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2016
Resident T Cells Are Unable To Control Herpes Simplex Virus-1 Activity in the Brain Ependymal Region during Latency.
Journal of immunology (Baltimore, Md. : 1950)
Menendez CM,Jinkins JK,Carr DJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Aug 15 00:00:00 EDT 2016
ECM-Dependence of Endothelial Progenitor Cell Features.
Journal of cellular biochemistry
Siavashi V,Nassiri SM,Rahbarghazi R,Vafaei R,Sariri R
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Effects of nerve growth factor and basic fibroblast growth factor dual gene modification on rat bone marrow mesenchymal stem cell differentiation into neuron-like cells in vitro.
Molecular medicine reports
Hu Y,Zhang Y,Tian K,Xun C,Wang S,Lv D
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2016
A flow cytometric approach to analyzing mature and progenitor endothelial cells following traumatic brain injury.
Journal of neuroscience methods
Assis-Nascimento P,Umland O,Cepero ML,Liebl DJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Apr 01 00:00:00 EDT 2016
A mouse model for a partially inactive obesity-associated human MC3R variant.
Nature communications
Lee B,Koo J,Yun Jun J,Gavrilova O,Lee Y,Seo AY,Taylor-Douglas DC,Adler-Wailes DC,Chen F,Gardner R,Koutzoumis D,Sherafat Kazemzadeh R,Roberson RB,Yanovski JA
15-0451 was used in Flow cytometry/Cell sorting to confirm the importance of MC3R signalling in human metabolism, and suggest a novel role for MC3R in adipose tissue development.
Thu Jan 28 00:00:00 EST 2016
A cell-autonomous tumour suppressor role of RAF1 in hepatocarcinogenesis.
Nature communications
Jeric I,Maurer G,Cavallo AL,Raguz J,Desideri E,Tarkowski B,Parrini M,Fischer I,Zatloukal K,Baccarini M
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Dec 21 00:00:00 EST 2016
Stromal senescence establishes an immunosuppressive microenvironment that drives tumorigenesis.
Nature communications
Ruhland MK,Loza AJ,Capietto AH,Luo X,Knolhoff BL,Flanagan KC,Belt BA,Alspach E,Leahy K,Luo J,Schaffer A,Edwards JR,Longmore G,Faccio R,DeNardo DG,Stewart SA
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Jun 08 00:00:00 EDT 2016
Targeting VEGF-A in myeloid cells enhances natural killer cell responses to chemotherapy and ameliorates cachexia.
Nature communications
Klose R,Krzywinska E,Castells M,Gotthardt D,Putz EM,Kantari-Mimoun C,Chikdene N,Meinecke AK,Schrödter K,Helfrich I,Fandrey J,Sexl V,Stockmann C
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Aug 19 00:00:00 EDT 2016
Protein Kinase D2 Protects against Acute Colitis Induced by Dextran Sulfate Sodium in Mice.
Scientific reports
Xiong J,Zhou MF,Wang YD,Chen LP,Xu WF,Wang YD,Deng F,Liu SD
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Sep 23 00:00:00 EDT 2016
Preparation of Single Cell Suspensions from Mouse Aorta.
Bio-protocol
Hu D,Yin C,Mohanta SK,Weber C,Habenicht AJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sun Jun 05 00:00:00 EDT 2016
Irradiation of necrotic cancer cells, employed for pulsing dendritic cells (DCs), potentiates DC vaccine-induced antitumor immunity against high-grade glioma.
Oncoimmunology
Vandenberk L,Garg AD,Verschuere T,Koks C,Belmans J,Beullens M,Agostinis P,De Vleeschouwer S,Van Gool SW
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2016
Diet-dependent, microbiota-independent regulation of IL-10-producing lamina propria macrophages in the small intestine.
Scientific reports
Ochi T,Feng Y,Kitamoto S,Nagao-Kitamoto H,Kuffa P,Atarashi K,Honda K,Teitelbaum DH,Kamada N
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Jun 15 00:00:00 EDT 2016
LTβR controls thymic portal endothelial cells for haematopoietic progenitor cell homing and T-cell regeneration.
Nature communications
Shi Y,Wu W,Chai Q,Li Q,Hou Y,Xia H,Ren B,Xu H,Guo X,Jin C,Lv M,Wang Z,Fu YX,Zhu M
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Aug 05 00:00:00 EDT 2016
Host microbiota constantly control maturation and function of microglia in the CNS.
Nature neuroscience
Erny D,Hrabě de Angelis AL,Jaitin D,Wieghofer P,Staszewski O,David E,Keren-Shaul H,Mahlakoiv T,Jakobshagen K,Buch T,Schwierzeck V,Utermöhlen O,Chun E,Garrett WS,McCoy KD,Diefenbach A,Staeheli P,Stecher B,Amit I,Prinz M
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2015
Vascular niche promotes hematopoietic multipotent progenitor formation from pluripotent stem cells.
The Journal of clinical investigation
Gori JL,Butler JM,Chan YY,Chandrasekaran D,Poulos MG,Ginsberg M,Nolan DJ,Elemento O,Wood BL,Adair JE,Rafii S,Kiem HP
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Mar 02 00:00:00 EST 2015
Tumor cells, but not endothelial cells, mediate eradication of primary sarcomas by stereotactic body radiation therapy.
Science translational medicine
Moding EJ,Castle KD,Perez BA,Oh P,Min HD,Norris H,Ma Y,Cardona DM,Lee CL,Kirsch DG
15-0451 was used in Flow cytometry/Cell sorting to investigate the contribution of different cell types to tumour response to radiation.
Wed Mar 11 00:00:00 EDT 2015
Phagocytosis-dependent activation of a TLR9-BTK-calcineurin-NFAT pathway co-ordinates innate immunity to Aspergillus fumigatus.
EMBO molecular medicine
Herbst S,Shah A,Mazon Moya M,Marzola V,Jensen B,Reed A,Birrell MA,Saijo S,Mostowy S,Shaunak S,Armstrong-James D
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2015
Tracking and Functional Characterization of Epithelial-Mesenchymal Transition and Mesenchymal Tumor Cells during Prostate Cancer Metastasis.
Cancer research
Ruscetti M,Quach B,Dadashian EL,Mulholland DJ,Wu H
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2015
Atypical chemokine receptor 1 deficiency reduces atherogenesis in ApoE-knockout mice.
Cardiovascular research
Wan W,Liu Q,Lionakis MS,Marino AP,Anderson SA,Swamydas M,Murphy PM
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2015
Dynamics and Transcriptomics of Skin Dendritic Cells and Macrophages in an Imiquimod-Induced, Biphasic Mouse Model of Psoriasis.
Journal of immunology (Baltimore, Md. : 1950)
Terhorst D,Chelbi R,Wohn C,Malosse C,Tamoutounour S,Jorquera A,Bajenoff M,Dalod M,Malissen B,Henri S
15-0451 was used in Flow cytometry/Cell sorting to develop and characterise an improved mouse model of psoriasis using imiquimod, showing that the dynamics and transcriptomics of skin dendritic cells and macrophages are similar to those seen in human pathology.
Sun Nov 15 00:00:00 EST 2015
Conditional immortalization of primary adipocyte precursor cells.
Adipocyte
Church C,Brown M,Rodeheffer MS
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Aug 10 00:00:00 EDT 2015
Nonadherent culture method downregulates stem cell antigen-1 expression in mouse bone marrow mesenchymal stem cells.
Experimental and therapeutic medicine
Deng B,Deng W,Xiao P,Zeng K,Zhang S,Zhang H,Deng DY,Yang Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2015
Favorable alteration of tumor microenvironment by immunomodulatory cytokines for efficient T-cell therapy in solid tumors.
PloS one
Tähtinen S,Kaikkonen S,Merisalo-Soikkeli M,Grönberg-Vähä-Koskela S,Kanerva A,Parviainen S,Vähä-Koskela M,Hemminki A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Apr 28 00:00:00 EDT 2016
The Nuclear Orphan Receptor NR2F6 Is a Central Checkpoint for Cancer Immune Surveillance.
Cell reports
Hermann-Kleiter N,Klepsch V,Wallner S,Siegmund K,Klepsch S,Tuzlak S,Villunger A,Kaminski S,Pfeifhofer-Obermair C,Gruber T,Wolf D,Baier G
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2015
Glioma-associated microglia/macrophages display an expression profile different from M1 and M2 polarization and highly express Gpnmb and Spp1.
PloS one
Szulzewsky F,Pelz A,Feng X,Synowitz M,Markovic D,Langmann T,Holtman IR,Wang X,Eggen BJ,Boddeke HW,Hambardzumyan D,Wolf SA,Kettenmann H
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2016
Targeted Gene Correction in Osteopetrotic-Induced Pluripotent Stem Cells for the Generation of Functional Osteoclasts.
Stem cell reports
Neri T,Muggeo S,Paulis M,Caldana ME,Crisafulli L,Strina D,Focarelli ML,Faggioli F,Recordati C,Scaramuzza S,Scanziani E,Mantero S,Buracchi C,Sobacchi C,Lombardo A,Naldini L,Vezzoni P,Villa A,Ficara F
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Oct 13 00:00:00 EDT 2015
Cardiac telocytes and fibroblasts in primary culture: different morphologies and immunophenotypes.
PloS one
Bei Y,Zhou Q,Fu S,Lv D,Chen P,Chen Y,Wang F,Xiao J
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Nov 11 00:00:00 EST 2015
Galectin-3 deficiency exacerbates hyperglycemia and the endothelial response to diabetes.
Cardiovascular diabetology
Darrow AL,Shohet RV
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Sat Jun 06 00:00:00 EDT 2015
Endothelial Gata5 transcription factor regulates blood pressure.
Nature communications
Messaoudi S,He Y,Gutsol A,Wight A,Hébert RL,Vilmundarson RO,Makrigiannis AP,Chalmers J,Hamet P,Tremblay J,McPherson R,Stewart AFR,Touyz RM,Nemer M
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Nov 30 00:00:00 EST 2015
Reprogramming human endothelial cells to haematopoietic cells requires vascular induction.
Nature
Sandler VM,Lis R,Liu Y,Kedem A,James D,Elemento O,Butler JM,Scandura JM,Rafii S
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Jul 17 00:00:00 EDT 2014
Developmental heterogeneity of cardiac fibroblasts does not predict pathological proliferation and activation.
Circulation research
Ali SR,Ranjbarvaziri S,Talkhabi M,Zhao P,Subat A,Hojjat A,Kamran P,Müller AM,Volz KS,Tang Z,Red-Horse K,Ardehali R
15-0451 was used in Flow cytometry/Cell sorting to study the developmental origins of cardiac fibroblasts and characterise their corresponding phenotypes, to aid in determining the proliferation rates of each developmental subset.
Fri Sep 12 00:00:00 EDT 2014
Atm deletion with dual recombinase technology preferentially radiosensitizes tumor endothelium.
The Journal of clinical investigation
Moding EJ,Lee CL,Castle KD,Oh P,Mao L,Zha S,Min HD,Ma Y,Das S,Kirsch DG
15-0451 was used in Flow cytometry/Cell sorting to determine that endothelial cells must progress through the cell cycle in order to be radiosensitized by Atm deletion.
Fri Aug 01 00:00:00 EDT 2014
Cell-autonomous heparanase modulates self-renewal and migration in bone marrow-derived mesenchymal stem cells.
Journal of biomedical science
Cheng CC,Lee YH,Lin SP,Huangfu WC,Liu IH
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Mar 13 00:00:00 EDT 2014
Astrocytic transforming growth factor-beta signaling reduces subacute neuroinflammation after stroke in mice.
Glia
Cekanaviciute E,Fathali N,Doyle KP,Williams AM,Han J,Buckwalter MS
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Aug 01 00:00:00 EDT 2014
Tracking the elusive fibrocyte: identification and characterization of collagen-producing hematopoietic lineage cells during murine wound healing.
Stem cells (Dayton, Ohio)
Suga H,Rennert RC,Rodrigues M,Sorkin M,Glotzbach JP,Januszyk M,Fujiwara T,Longaker MT,Gurtner GC
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu May 01 00:00:00 EDT 2014
Bone marrow mesenchymal stem cells provide an antibiotic-protective niche for persistent viable Mycobacterium tuberculosis that survive antibiotic treatment.
The American journal of pathology
Beamer G,Major S,Das B,Campos-Neto A
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Dec 01 00:00:00 EST 2014
Caveolin-1 increases proinflammatory chemoattractants and blood-retinal barrier breakdown but decreases leukocyte recruitment in inflammation.
Investigative ophthalmology & visual science
Li X,Gu X,Boyce TM,Zheng M,Reagan AM,Qi H,Mandal N,Cohen AW,Callegan MC,Carr DJ,Elliott MH
15-0451 was used in Flow cytometry/Cell sorting to examine the effect of Caveolin-1 ablation on the sensitivity of the retina to inflammation.
Tue Aug 26 00:00:00 EDT 2014
Corneal inflammation after miniature keratoprosthesis implantation.
Investigative ophthalmology & visual science
Crnej A,Omoto M,Dohlman TH,Dohlman CH,Dana R
15-0451 was used in Flow cytometry/Cell sorting to suggest that using syngeneic or decellularised corneal tissue as a Boston-KPro carrier could reduce the postoperative inflammation response.
Tue Dec 16 00:00:00 EST 2014
Identification of hematopoietic-specific regulatory elements from the CD45 gene and use for lentiviral tracking of transplanted cells.
Experimental hematology
Duong KL,Das S,Yu S,Barr JY,Jena S,Kim E,Zavazava N,Colgan JD,Xue HH,Levasseur DN
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
A new animal model of spontaneous autoimmune peripheral polyneuropathy: implications for Guillain-Barré syndrome.
Acta neuropathologica communications
Yang M,Rainone A,Shi XQ,Fournier S,Zhang J
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Jan 08 00:00:00 EST 2014
Amniotic fluid stem cells prevent follicle atresia and rescue fertility of mice with premature ovarian failure induced by chemotherapy.
PloS one
Xiao GY,Liu IH,Cheng CC,Chang CC,Lee YH,Cheng WT,Wu SC
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu May 28 00:00:00 EDT 2015
Activation of NLRP3 inflammasome by crystalline structures via cell surface contact.
Scientific reports
Hari A,Zhang Y,Tu Z,Detampel P,Stenner M,Ganguly A,Shi Y
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Dec 02 00:00:00 EST 2014
The nucleotide-binding leucine-rich repeat (NLR) family member NLRX1 mediates protection against experimental autoimmune encephalomyelitis and represses macrophage/microglia-induced inflammation.
The Journal of biological chemistry
Eitas TK,Chou WC,Wen H,Gris D,Robbins GR,Brickey J,Oyama Y,Ting JP
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Fri Feb 14 00:00:00 EST 2014
Isolation, characterization, and transplantation of cardiac endothelial cells.
BioMed research international
Pratumvinit B,Reesukumal K,Janebodin K,Ieronimakis N,Reyes M
15-0451 was used in Flow cytometry/Cell sorting to develop a method to expand the study of endothelial cell biology for multiple clinical applications..
Tue Jun 17 00:00:00 EDT 2014
Methotrexate treatment causes early onset of disease in a mouse model of Ross River virus-induced inflammatory disease through increased monocyte production.
PloS one
Taylor A,Sheng KC,Herrero LJ,Chen W,Rulli NE,Mahalingam S
15-0451 was used in Flow cytometry/Cell sorting to study infiltration of inflammatory cells to skeletal muscle in Ross River virus disease in response to MTX treatment.
Thu Mar 06 00:00:00 EST 2014
Gli1 deletion prevents Helicobacter-induced gastric metaplasia and expansion of myeloid cell subsets.
PloS one
El-Zaatari M,Kao JY,Tessier A,Bai L,Hayes MM,Fontaine C,Eaton KA,Merchant JL
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Sep 10 00:00:00 EDT 2013
Tumorigenic cells are common in mouse MPNSTs but their frequency depends upon tumor genotype and assay conditions.
Cancer cell
Buchstaller J,McKeever PE,Morrison SJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Feb 14 00:00:00 EST 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2012
A model of GAG/MIP-2/CXCR2 interfaces and its functional effects.
Biochemistry
Rajasekaran D,Keeler C,Syed MA,Jones MC,Harrison JK,Wu D,Bhandari V,Hodsdon ME,Lolis EJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Jul 17 00:00:00 EDT 2012
ER71 directs mesodermal fate decisions during embryogenesis.
Development (Cambridge, England)
Rasmussen TL,Kweon J,Diekmann MA,Belema-Bedada F,Song Q,Bowlin K,Shi X,Ferdous A,Li T,Kyba M,Metzger JM,Koyano-Nakagawa N,Garry DJ
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2011
Effect of topical azithromycin on corneal innate immune responses.
Investigative ophthalmology & visual science
Sadrai Z,Hajrasouliha AR,Chauhan S,Saban DR,Dastjerdi MH,Dana R
15-0451 was used in Flow cytometry/Cell sorting to suggest topical azithromycin may have an anti-inflammatory effect on corneal inflammation in a murine model.
Tue Apr 19 00:00:00 EDT 2011
Effect of topical azithromycin on corneal innate immune responses.
Investigative ophthalmology & visual science
Sadrai Z,Hajrasouliha AR,Chauhan S,Saban DR,Dastjerdi MH,Dana R
15-0451 was used in Flow cytometry/Cell sorting to suggest topical azithromycin may have an anti-inflammatory effect on corneal inflammation in a murine model.
Tue Apr 19 00:00:00 EDT 2011
Myeloid cells migrate in response to IL-24.
Cytokine
Buzas K,Oppenheim JJ,Zack Howard OM
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Thu Sep 01 00:00:00 EDT 2011
Lack of iNKT cells in patients with combined immune deficiency due to hypomorphic RAG mutations.
Blood
Matangkasombut P,Pichavant M,Saez DE,Giliani S,Mazzolari E,Finocchi A,Villa A,Sobacchi C,Cortes P,Umetsu DT,Notarangelo LD
15-0451 was used in Flow cytometry/Cell sorting to show that the absence of Invariant natural killer T cells in the peripheral blood of patients with hypomorphic RAG mutations, may contribute to the physical pathology of Ommen syndrome.
Tue Jan 01 00:00:00 EST 2008
Very small embryonic-like stem cells are present in adult murine organs: ImageStream-based morphological analysis and distribution studies.
Cytometry. Part A : the journal of the International Society for Analytical Cytology
Zuba-Surma EK,Kucia M,Wu W,Klich I,Lillard JW,Ratajczak J,Ratajczak MZ
15-0451 was used in Flow cytometry/Cell sorting to identify a population of primitive cells expressing germ line/epiblast markers deposited early during embryogenesis in various organs and survives into adulthood.
Mon Dec 01 00:00:00 EST 2008
Optimized isolation enables ex vivo analysis of microglia from various central nervous system regions.
Glia
de Haas AH,Boddeke HW,Brouwer N,Biber K
Published figure using CD45 monoclonal antibody (Product # 15-0451-82) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2007
Selective labeling and isolation of functional classes of interstitial cells of Cajal of human and murine small intestine.
American journal of physiology. Cell physiology
Chen H,Redelman D,Ro S,Ward SM,Ordög T,Sanders KM
15-0451 was used in Flow cytometry/Cell sorting to investigate different functional classes of interstitial cells of Cajal of human and murine small intestine.
Mon Jan 01 00:00:00 EST 2007