Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 5
Description: The HIS24 monoclonal antibody reacts with an isoform of CD45 expressed by rat B cells. The antigen recognized by HIS24 is found on B cells from the early stage of B cell development in the bone marrow to mature peripheral B cells, but not on plasma cells. HIS24 recognizes a 205 kDa protein by immunoblotting. CD45, also known as Leukocyte Common Antigen (LCA), has tyrosine phosphatase enzymatic activity in its cytoplasmic portion and plays an important role in activation of lymphocytes.
Applications Reported: This HIS24 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections.
Applications Tested: This HIS24 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. This HIS24 antibody has been tested by immunohistochemistry of frozen rat tissue and can be used at less than or equal to 10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.
Filtration: 0.2 µm post-manufacturing filtered.
CD45R (PTPRC) is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. CD45 contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains. CD45 is specifically expressed in hematopoietic cells and has been shown to be an essential regulator of T- and B-cell antigen receptor signaling. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists in multiple isoforms as a result of alternative splicing, and these isoforms differ in their extracellular domains, but share identical transmembrane and cytoplasmic domains. Further, these CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. In addition to its role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. Diseases associated with CD45 dysfunction include Severe Combined Immunodeficiency (T-cell negative, B-Cell Negative) and Hepatitis C.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD45; L-CA; leucocyte common antigen; Leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; Protein tyrosine phosphatase, receptor-type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein
Gene Aliases: CD45; L-CA; Lca; Ptprc; RT7; T200
UniProt ID: (Rat) P04157
Entrez Gene ID: (Rat) 24699
Molecular Function:
extracellular matrix protein
protein phosphatase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support