Product References
Nucleolar protein TAAP1/C22orf46 confers pro-survival signaling in non-small cell lung cancer.
Life science alliance
Döring M,Brux M,Paszkowski-Rogacz M,Guillem-Gloria PM,Buchholz F,Pisabarro MT,Theis M
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Mon Apr 01 00:00:00 UTC 2024
Anti-CD47 Antibody Enhances the Efficacy of Chemotherapy in Patients with Gastric Cancer Liver Metastasis.
Journal of Cancer
Liu Z,Chen H,Ta N,Shi Z,Zhan L,Han T,Zhang J,Chang X,Yin K,Nie M
14-0479-82 was used in Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, In Vivo to demonstrate that tumor-derived exosomes are involved in GCLM progression, targeting CD47 inhibits gastric cancer tumorigenesis, and a combination of anti-CD47 antibodies and 5-Fu shows potential for treating GCLM.
Fri Mar 03 00:00:00 UTC 2023
Altered Membrane Expression and Function of CD11b Play a Role in the Immunosuppressive Effects of Morphine on Macrophages at the Nanomolar Level.
Pharmaceuticals (Basel, Switzerland)
Yu PC,Hao CY,Fan YZ,Liu D,Qiao YF,Yao JB,Li CZ,Yu Y
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Mon Feb 13 00:00:00 UTC 2023
Phagocytosis-initiated tumor hybrid cells acquire a c-Myc-mediated quasi-polarization state for immunoevasion and distant dissemination.
Nature communications
Chou CW,Hung CN,Chiu CH,Tan X,Chen M,Chen CC,Saeed M,Hsu CW,Liss MA,Wang CM,Lai Z,Alvarez N,Osmulski PA,Gaczynska ME,Lin LL,Ortega V,Kirma NB,Xu K,Liu Z,Kumar AP,Taverna JA,Velagaleti GVN,Chen CL,Zhang Z,Huang TH
14-0479-82 was used in Flow Cytometry to report that the reverse signaling using CD47 as a receptor additionally enhances a pro-survival function of prostate cancer cells under phagocytic attack.
Tue Oct 17 00:00:00 UTC 2023
Melanophages give rise to hyperreflective foci in AMD, a disease-progression marker.
Journal of neuroinflammation
Augustin S,Lam M,Lavalette S,Verschueren A,Blond F,Forster V,Przegralek L,He Z,Lewandowski D,Bemelmans AP,Picaud S,Sahel JA,Mathis T,Paques M,Thuret G,Guillonneau X,Delarasse C,Sennlaub F
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Inhibition Assays
Wed Feb 08 00:00:00 UTC 2023
Melanophages give rise to hyperreflective foci in AMD, a disease-progression marker.
Journal of neuroinflammation
Augustin S,Lam M,Lavalette S,Verschueren A,Blond F,Forster V,Przegralek L,He Z,Lewandowski D,Bemelmans AP,Picaud S,Sahel JA,Mathis T,Paques M,Thuret G,Guillonneau X,Delarasse C,Sennlaub F
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Inhibition Assays
Wed Feb 08 00:00:00 UTC 2023
CD47 Blockade Accelerates Blood Clearance and Alleviates Early Brain Injury After Experimental Subarachnoid Hemorrhage.
Frontiers in immunology
Xu CR,Li JR,Jiang SW,Wan L,Zhang X,Xia L,Hua XM,Li ST,Chen HJ,Fu XJ,Jing CH
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Inhibition Assays
Mon May 02 00:00:00 UTC 2022
Schweinfurthin induces ICD without ER stress and caspase activation.
Oncoimmunology
Zhang R,Neighbors JD,Schell TD,Hohl RJ
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Tue Aug 09 00:00:00 UTC 2022
Anti-CD47 antibody synergizes with cisplatin against laryngeal cancer by enhancing phagocytic ability of macrophages.
Clinical and experimental immunology
Wang J,Zhang H,Yin X,Bian Y
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Wed Sep 01 00:00:00 UTC 2021
LSD1 silencing contributes to enhanced efficacy of anti-CD47/PD-L1 immunotherapy in cervical cancer.
Cell death & disease
Xu S,Wang X,Yang Y,Li Y,Wu S
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Wed Mar 17 00:00:00 UTC 2021
Exosome-mediated delivery of CRISPR/Cas9 for targeting of oncogenic KrasG12D in pancreatic cancer.
Life science alliance
McAndrews KM,Xiao F,Chronopoulos A,LeBleu VS,Kugeratski FG,Kalluri R
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Wed Sep 01 00:00:00 UTC 2021
Quantitative proteomics identifies the core proteome of exosomes with syntenin-1 as the highest abundant protein and a putative universal biomarker.
Nature cell biology
Kugeratski FG,Hodge K,Lilla S,McAndrews KM,Zhou X,Hwang RF,Zanivan S,Kalluri R
14-0479-82 was used in Flow cytometry/Cell sorting to provide a comprehensive quantitative atlas of core proteins ubiquitous to exosomes that can serve as a resource for the scientific community.
Tue Jun 01 00:00:00 UTC 2021
CD47 prevents the elimination of diseased fibroblasts in scleroderma.
JCI insight
Lerbs T,Cui L,King ME,Chai T,Muscat C,Chung L,Brown R,Rieger K,Shibata T,Wernig G
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Immunocytochemistry
Thu Aug 20 00:00:00 UTC 2020
CD47 prevents the elimination of diseased fibroblasts in scleroderma.
JCI insight
Lerbs T,Cui L,King ME,Chai T,Muscat C,Chung L,Brown R,Rieger K,Shibata T,Wernig G
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Immunocytochemistry
Thu Aug 20 00:00:00 UTC 2020
Therapeutic modulation of phagocytosis in glioblastoma can activate both innate and adaptive antitumour immunity.
Nature communications
von Roemeling CA,Wang Y,Qie Y,Yuan H,Zhao H,Liu X,Yang Z,Yang M,Deng W,Bruno KA,Chan CK,Lee AS,Rosenfeld SS,Yun K,Johnson AJ,Mitchell DA,Jiang W,Kim BYS
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Immunohistochemistry
Fri Mar 20 00:00:00 UTC 2020
Therapeutic modulation of phagocytosis in glioblastoma can activate both innate and adaptive antitumour immunity.
Nature communications
von Roemeling CA,Wang Y,Qie Y,Yuan H,Zhao H,Liu X,Yang Z,Yang M,Deng W,Bruno KA,Chan CK,Lee AS,Rosenfeld SS,Yun K,Johnson AJ,Mitchell DA,Jiang W,Kim BYS
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Immunohistochemistry
Fri Mar 20 00:00:00 UTC 2020
Regulation of senescence escape by TSP1 and CD47 following chemotherapy treatment.
Cell death & disease
Guillon J,Petit C,Moreau M,Toutain B,Henry C,Roché H,Bonichon-Lamichhane N,Salmon JP,Lemonnier J,Campone M,Verrièle V,Lelièvre E,Guette C,Coqueret O
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Wed Feb 27 00:00:00 UTC 2019
Regulation of senescence escape by TSP1 and CD47 following chemotherapy treatment.
Cell death & disease
Guillon J,Petit C,Moreau M,Toutain B,Henry C,Roché H,Bonichon-Lamichhane N,Salmon JP,Lemonnier J,Campone M,Verrièle V,Lelièvre E,Guette C,Coqueret O
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Wed Feb 27 00:00:00 UTC 2019
Regulation of senescence escape by TSP1 and CD47 following chemotherapy treatment.
Cell death & disease
Guillon J,Petit C,Moreau M,Toutain B,Henry C,Roché H,Bonichon-Lamichhane N,Salmon JP,Lemonnier J,Campone M,Verrièle V,Lelièvre E,Guette C,Coqueret O
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Wed Feb 27 00:00:00 UTC 2019
Generation and testing of clinical-grade exosomes for pancreatic cancer.
JCI insight
Mendt M,Kamerkar S,Sugimoto H,McAndrews KM,Wu CC,Gagea M,Yang S,Blanko EVR,Peng Q,Ma X,Marszalek JR,Maitra A,Yee C,Rezvani K,Shpall E,LeBleu VS,Kalluri R
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Thu Apr 19 00:00:00 UTC 2018
Exosomes facilitate therapeutic targeting of oncogenic KRAS in pancreatic cancer.
Nature
Kamerkar S,LeBleu VS,Sugimoto H,Yang S,Ruivo CF,Melo SA,Lee JJ,Kalluri R
14-0479 was used in Flow cytometry/Cell sorting to develop a therapeutic strategy for targeting mutant KRAS in pancreatic cancer using exosomes carrying specific short interfering or short hairpin RNA.
Thu Jun 22 00:00:00 UTC 2017
CD47 promotes ovarian cancer progression by inhibiting macrophage phagocytosis.
Oncotarget
Liu R,Wei H,Gao P,Yu H,Wang K,Fu Z,Ju B,Zhao M,Dong S,Li Z,He Y,Huang Y,Yao Z
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Tue Jun 13 00:00:00 UTC 2017
CD47 promotes ovarian cancer progression by inhibiting macrophage phagocytosis.
Oncotarget
Liu R,Wei H,Gao P,Yu H,Wang K,Fu Z,Ju B,Zhao M,Dong S,Li Z,He Y,Huang Y,Yao Z
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Tue Jun 13 00:00:00 UTC 2017
SIRPα-antibody fusion proteins stimulate phagocytosis and promote elimination of acute myeloid leukemia cells.
Oncotarget
Ponce LP,Fenn NC,Moritz N,Krupka C,Kozik JH,Lauber K,Subklewe M,Hopfner KP
14-0479 was used in Flow cytometry/Cell sorting to qualify local inhibitory checkpoint monoclonal antibodies as promising approach to confine the benefit of disrupting the CD47-SIRPα axis to tumor antigen-expressing cells.
Tue Feb 14 00:00:00 UTC 2017
Nanoparticle biointerfacing by platelet membrane cloaking.
Nature
Hu CM,Fang RH,Wang KC,Luk BT,Thamphiwatana S,Dehaini D,Nguyen P,Angsantikul P,Wen CH,Kroll AV,Carpenter C,Ramesh M,Qu V,Patel SH,Zhu J,Shi W,Hofman FM,Chen TC,Gao W,Zhang K,Chien S,Zhang L
14-0479 was used in Flow cytometry/Cell sorting to explore the use of the preparation of polymeric nanoparticles enclosed in the plasma membrane of human platelets.
Thu Oct 01 00:00:00 UTC 2015
CD47 is an adverse prognostic factor and a therapeutic target in gastric cancer.
Cancer medicine
Yoshida K,Tsujimoto H,Matsumura K,Kinoshita M,Takahata R,Matsumoto Y,Hiraki S,Ono S,Seki S,Yamamoto J,Hase K
14-0479 was used in Flow cytometry/Cell sorting to demonstrate that higher expressions of CD44 and CD47 in gastric cancer cells correlate with higher proliferation and tumorigenicity.
Tue Sep 01 00:00:00 UTC 2015
Exploiting light chains for the scalable generation and platform purification of native human bispecific IgG.
Nature communications
Fischer N,Elson G,Magistrelli G,Dheilly E,Fouque N,Laurendon A,Gueneau F,Ravn U,Depoisier JF,Moine V,Raimondi S,Malinge P,Di Grazia L,Rousseau F,Poitevin Y,Calloud S,Cayatte PA,Alcoz M,Pontini G,Fagète S,Broyer L,Corbier M,Schrag D,Didelot G,Bosson N,Costes N,Cons L,Buatois V,Johnson Z,Ferlin W,Masternak K,Kosco-Vilbois M
14-0479 was used in Flow cytometry/Cell sorting to demonstrate that light chains can play a dominant role in mediating specificity and high affinity.
Thu Feb 12 00:00:00 UTC 2015
Cells expressing the C/EBPbeta isoform, LIP, engulf their neighbors.
PloS one
Abreu M,Sealy L
Published figure using CD47 monoclonal antibody (Product # 14-0479-82) in Flow Cytometry
Thu Apr 11 00:00:00 UTC 2013
CD14 and toll-like receptors 2 and 4 are required for fibrillar A{beta}-stimulated microglial activation.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Reed-Geaghan EG,Savage JC,Hise AG,Landreth GE
14-0479 was used in Blocking experiments to study how the engagement of the fAbeta receptor complex is linked to the induction of an activated microglial phenotype.
Wed Sep 23 00:00:00 UTC 2009