Product References
Perinatal murine cytomegalovirus infection reshapes the transcriptional profile and functionality of NK cells.
Nature communications
Rožmanić C,Lisnić B,Pribanić Matešić M,Mihalić A,Hiršl L,Park E,Lesac Brizić A,Indenbirken D,Viduka I,Šantić M,Adler B,Yokoyama WM,Krmpotić A,Juranić Lisnić V,Jonjić S,Brizić I
Published figure using CD49b (Integrin alpha 2) monoclonal antibody (Product # 17-5971-82) in Flow Cytometry
Thu Oct 12 00:00:00 UTC 2023
Stage-specific action of Runx1 and GATA3 controls silencing of PU.1 expression in mouse pro-T cells.
The Journal of experimental medicine
Hosokawa H,Koizumi M,Masuhara K,Romero-Wolf M,Tanaka T,Nakayama T,Rothenberg EV
Published figure using CD49b (Integrin alpha 2) monoclonal antibody (Product # 17-5971-82) in Flow Cytometry
Mon Aug 02 00:00:00 UTC 2021
Myeloid cell subsets that express latency-associated peptide promote cancer growth by modulating T cells.
iScience
Gabriely G,Ma D,Siddiqui S,Sun L,Skillin NP,Abou-El-Hassan H,Moreira TG,Donnelly D,da Cunha AP,Fujiwara M,Walton LR,Patel A,Krishnan R,Levine SS,Healy BC,Rezende RM,Murugaiyan G,Weiner HL
17-5971-82 was used in Flow cytometry/Cell sorting to identify a previously unknown function by which LAPHi MCs promote tumor growth and offer therapeutic intervention to target these cells in cancer.
Fri Nov 19 00:00:00 UTC 2021
Dual vaccination against IL-4 and IL-13 protects against chronic allergic asthma in mice.
Nature communications
Conde E,Bertrand R,Balbino B,Bonnefoy J,Stackowicz J,Caillot N,Colaone F,Hamdi S,Houmadi R,Loste A,Kamphuis JBJ,Huetz F,Guilleminault L,Gaudenzio N,Mougel A,Hardy D,Snouwaert JN,Koller BH,Serra V,Bruhns P,Grouard-Vogel G,Reber LL
17-5971-82 was used in Flow Cytometry to imply that dual IL-4/IL-13 vaccination may represent a cost-effective, long-term therapeutic strategy for the treatment of allergic asthma as demonstrated in mouse models, although additional studies are warranted to assess its safety and feasibility.
Tue May 11 00:00:00 UTC 2021
GM-CSF production by non-classical monocytes controls antagonistic LPS-driven functions in allergic inflammation.
Cell reports
Kaur K,Bachus H,Lewis C,Papillion AM,Rosenberg AF,Ballesteros-Tato A,León B
17-5971-82 was used in Flow cytometry/Cell sorting to show here that lipopolysaccharide activity switches from pro-pathogenic to protective depending on the production of granulocyte-macrophage colony-stimulating factor by non-classical monocytes.
Tue Dec 28 00:00:00 UTC 2021
Stage-Specific Requirement for Eomes in Mature NK Cell Homeostasis and Cytotoxicity.
Cell reports
Wagner JA,Wong P,Schappe T,Berrien-Elliott MM,Cubitt C,Jaeger N,Lee M,Keppel CR,Marin ND,Foltz JA,Marsala L,Neal CC,Sullivan RP,Schneider SE,Keppel MP,Saucier N,Cooper MA,Fehniger TA
17-5971 was used in Flow cytometry/Cell sorting to develop a tamoxifen-inducible, type-1-ILC-specific (Ncr1-targeted) cre mouse and combine this with Eomes-floxed mice.
Tue Jun 02 00:00:00 UTC 2020
Natural Killer Cells Degenerate Intact Sensory Afferents following Nerve Injury.
Cell
Davies AJ,Kim HW,Gonzalez-Cano R,Choi J,Back SK,Roh SE,Johnson E,Gabriac M,Kim MS,Lee J,Lee JE,Kim YS,Bae YC,Kim SJ,Lee KM,Na HS,Riva P,Latremoliere A,Rinaldi S,Ugolini S,Costigan M,Oh SB
17-5971 was used in Flow cytometry/Cell sorting to suggest the therapeutic potential of modulating NK cell function to resolve painful neuropathy through the clearance of partially damaged nerves.
Thu Feb 07 00:00:00 UTC 2019
Polychromic Reporter Mice Reveal Unappreciated Innate Lymphoid Cell Progenitor Heterogeneity and Elusive ILC3 Progenitors in Bone Marrow.
Immunity
Walker JA,Clark PA,Crisp A,Barlow JL,Szeto A,Ferreira ACF,Rana BMJ,Jolin HE,Rodriguez-Rodriguez N,Sivasubramaniam M,Pannell R,Cruickshank J,Daly M,Haim-Vilmovsky L,Teichmann SA,McKenzie ANJ
17-5971 was used in Flow cytometry/Cell sorting to identify a discrete ILC2-committed population and delineated transition states between early progenitors and a highly heterogeneous ILC1, ILC3, and NK precursor cell cluster.
Tue Jul 16 00:00:00 UTC 2019
Progranulin prevents regulatory NK cell cytotoxicity against antiviral T cells.
JCI insight
Huang A,Shinde PV,Huang J,Senff T,Xu HC,Margotta C,Häussinger D,Willnow TE,Zhang J,Pandyra AA,Timm J,Weggen S,Lang KS,Lang PA
17-5971 was used in Flow cytometry/Cell sorting to show that progranulin (PGRN) can reduce NK cell cytotoxicity through reduction of NK cell expansion, granzyme B transcription, and NK cell-mediated lysis of target cells.
Thu Sep 05 00:00:00 UTC 2019
Chemo-photothermal therapy combination elicits anti-tumor immunity against advanced metastatic cancer.
Nature communications
Nam J,Son S,Ochyl LJ,Kuai R,Schwendeman A,Moon JJ
17-5971 was used in Flow cytometry/Cell sorting to show that chemo-photothermal therapy tuned to elicit anti-tumour immunity can exert striking therapeutic effects against primary and disseminated tumours.
Wed Mar 14 00:00:00 UTC 2018
Investigation of tumor-tumor interactions in a double human cervical carcinoma xenograft model in nude mice.
Oncotarget
Mertens B,Cristina de Araujo Nogueira T,Topalis D,Stranska R,Snoeck R,Andrei G
17-5971 was used in Flow cytometry/Cell sorting to identify the potential use of cidofovir in therapeutic strategies to kill tumour cells and influence the immune system to fight cancer.
Tue Apr 24 00:00:00 UTC 2018
Spontaneous Chitin Accumulation in Airways and Age-Related Fibrotic Lung Disease.
Cell
Van Dyken SJ,Liang HE,Naikawadi RP,Woodruff PG,Wolters PJ,Erle DJ,Locksley RM
17-5971 was used in Flow cytometry/Cell sorting to show that altered chitin clearance could exacerbate fibrogenic pathways in the setting of lung diseases characterised by epithelial cell dysfunction.
Thu Apr 20 00:00:00 UTC 2017
Preferential TNFα signaling via TNFR2 regulates epithelial injury and duct obstruction in experimental biliary atresia.
JCI insight
Shivakumar P,Mizuochi T,Mourya R,Gutta S,Yang L,Luo Z,Bezerra JA
17-5971 was used in Magnetic cell separation to determine factors regulating the pathogenic mechanisms of biliary atresia.
Thu Mar 09 00:00:00 UTC 2017
The breast tumor microenvironment alters the phenotype and function of natural killer cells.
Cellular & molecular immunology
Krneta T,Gillgrass A,Chew M,Ashkar AA
17-5971 was used in Flow cytometry/Cell sorting to investigate the developmental stage of tumour-associated NK cells, showing that the tumour microenvironment alters the phenotype and function of NK cells.
Thu Sep 01 00:00:00 UTC 2016
IDO1 Deficiency Does Not Affect Disease in Mouse Models of Systemic Juvenile Idiopathic Arthritis and Secondary Hemophagocytic Lymphohistiocytosis.
PloS one
Put K,Brisse E,Avau A,Imbrechts M,Mitera T,Janssens R,Proost P,Fallarino F,Wouters CH,Matthys P
17-5971 was used in Flow cytometry/Cell sorting to unravel the role of IDO1, an immune-modulatory enzyme, in the pathogenesis of certain inflammatory diseases.
Mon Aug 08 00:00:00 UTC 2016
A role for multiple chimeric antigen receptor-expressing leukocytes in antigen-specific responses to cancer.
Oncotarget
Yong CS,John LB,Devaud C,Prince MH,Johnstone RW,Trapani JA,Darcy PK,Kershaw MH
17-5971 was used in Flow cytometry/Cell sorting to characterise the function of leukocytes from transgenic mice expressing a chimeric antigen receptor under the control of vav, and determine the ability of these mice to respond to ERB-expressing tumours.
Tue Jun 07 00:00:00 UTC 2016
Intratumoral INF-γ triggers an antiviral state in GL261 tumor cells: a major hurdle to overcome for oncolytic vaccinia virus therapy of cancer.
Molecular therapy oncolytics
Kober C,Weibel S,Rohn S,Kirscher L,Szalay AA
17-5971 was used in Flow cytometry/Cell sorting to identify microenvironmental factors, which may be decisive for failure or success of oncolytic VACV therapy.
Wed Apr 27 00:00:00 UTC 2016
Natural killer cells promote long-term hepatobiliary inflammation in a low-dose rotavirus model of experimental biliary atresia.
PloS one
Squires JE,Shivakumar P,Mourya R,Bessho K,Walters S,Bezerra JA
17-5971 was used in Flow cytometry/Cell sorting to investigate mechanisms of liver injury in a mouse model of Rhesus rotavirus-induced biliary atresia.
Fri Apr 15 00:00:00 UTC 2016
Expression of a Chimeric Antigen Receptor in Multiple Leukocyte Lineages in Transgenic Mice.
PloS one
Yong CS,Westwood JA,Schröder J,Papenfuss AT,von Scheidt B,Moeller M,Devaud C,Darcy PK,Kershaw MH
17-5971 was used in Flow cytometry/Cell sorting to generate a mouse model expressing chimeric antigen receptors (CARs) specific for a tumour antigen under a pan-hematopoietic promoter.
Wed Jun 08 00:00:00 UTC 2016
IFN-γ modulates Ly-49 receptors on NK cells in IFN-γ-induced pregnancy failure.
Scientific reports
Li ZY,Song ZH,Meng CY,Yang DD,Yang Y,Peng JP
17-5971 was used in Flow cytometry/Cell sorting to investigate the mechanism by which IFN-γ modulates Ly-49 receptors on natural killer cells during pregnancy failure.
Fri Dec 11 00:00:00 UTC 2015
Biliary repair and carcinogenesis are mediated by IL-33-dependent cholangiocyte proliferation.
The Journal of clinical investigation
Li J,Razumilava N,Gores GJ,Walters S,Mizuochi T,Mourya R,Bessho K,Wang YH,Glaser SS,Shivakumar P,Bezerra JA
17-5971 was used in Flow cytometry/Cell sorting to demonstrate the role of the Th2-promoting cytokine, IL-33, in cholangiocyte proliferation.
Tue Jul 01 00:00:00 UTC 2014
Targeting CD137 enhances the efficacy of cetuximab.
The Journal of clinical investigation
Kohrt HE,Colevas AD,Houot R,Weiskopf K,Goldstein MJ,Lund P,Mueller A,Sagiv-Barfi I,Marabelle A,Lira R,Troutner E,Richards L,Rajapaska A,Hebb J,Chester C,Waller E,Ostashko A,Weng WK,Chen L,Czerwinski D,Fu YX,Sunwoo J,Levy R
17-5971 was used in Flow cytometry/Cell sorting to study the effects of targeting CD137 on modulating the anti-cancer efficacy of cetuximab.
Sun Jun 01 00:00:00 UTC 2014
Contribution of mast cell-derived interleukin-1β to uric acid crystal-induced acute arthritis in mice.
Arthritis & rheumatology (Hoboken, N.J.)
Reber LL,Marichal T,Sokolove J,Starkl P,Gaudenzio N,Iwakura Y,Karasuyama H,Schwartz LB,Robinson WH,Tsai M,Galli SJ
17-5971 was used in Flow cytometry/Cell sorting to assess the role of mast cells in a mouse model of monosodium urate monohydrate crystal-induced acute arthritis.
Wed Oct 01 00:00:00 UTC 2014
Signaling pathways activated by a protease allergen in basophils.
Proceedings of the National Academy of Sciences of the United States of America
Rosenstein RK,Bezbradica JS,Yu S,Medzhitov R
17-5971 was used in Flow cytometry/Cell sorting to study signaling pathways activated by a protease allergen in basophils.
Tue Nov 18 00:00:00 UTC 2014
Expression of a single ICAM-1 isoform on T cells is sufficient for development of experimental autoimmune encephalomyelitis.
European journal of immunology
Bullard DC,Hu X,Crawford D,McDonald K,Ramos TN,Barnum SR
17-5971 was used in Flow cytometry/Cell sorting to assess the role of a particular ICAM-1 isoform expressed by T-cells in the development of of experimental autoimmune encephalomyelitis.
Tue Apr 01 00:00:00 UTC 2014
An ENU-induced splicing mutation reveals a role for Unc93b1 in early immune cell activation following influenza A H1N1 infection.
Genes and immunity
Lafferty EI,Flaczyk A,Angers I,Homer R,d'Hennezel E,Malo D,Piccirillo CA,Vidal SM,Qureshi ST
17-5971 was used in Flow cytometry/Cell sorting to establish a role for Unc93b1 and endosomal TLRs in the activation of both myeloid and lymphoid cells during the innate immune response to influenza.
Fri Apr 10 00:00:00 UTC 2015
Core needle biopsy of breast cancer tumors increases distant metastases in a mouse model.
Neoplasia (New York, N.Y.)
Mathenge EG,Dean CA,Clements D,Vaghar-Kashani A,Photopoulos S,Coyle KM,Giacomantonio M,Malueth B,Nunokawa A,Jordan J,Lewis JD,Gujar SA,Marcato P,Lee PW,Giacomantonio CA
17-5971 was used in Flow cytometry/Cell sorting to demonstrate that CNB creates an immunosuppressive tumour microenvironment, increases EMT, and facilitates release of circulating tumour cells, leading to increased metastases.
Sat Nov 01 00:00:00 UTC 2014
Allogeneic splenocyte transfer and lipopolysaccharide inhalations induce differential T cell expansion and lung injury: a novel model of pulmonary graft-versus-host disease.
PloS one
Martinu T,Kinnier CV,Sun J,Kelly FL,Nelson ME,Garantziotis S,Foster WM,Palmer SM
17-5971 was used in Flow cytometry/Cell sorting to provide a novel model to dissect mechanisms of pGVHD independent of conditioning.
Thu Jan 15 00:00:00 UTC 2015
Expression of the secondary granule proteins major basic protein 1 (MBP-1) and eosinophil peroxidase (EPX) is required for eosinophilopoiesis in mice.
Blood
Doyle AD,Jacobsen EA,Ochkur SI,McGarry MP,Shim KG,Nguyen DT,Protheroe C,Colbert D,Kloeber J,Neely J,Shim KP,Dyer KD,Rosenberg HF,Lee JJ,Lee NA
17-5971 was used in Flow cytometry/Cell sorting to investigate the contributions of the granule proteins MBP-1 and EPX to eosinophil activity using double knockout mice.
Thu Aug 01 00:00:00 UTC 2013
Homologous recombination into the eosinophil peroxidase locus generates a strain of mice expressing Cre recombinase exclusively in eosinophils.
Journal of leukocyte biology
Doyle AD,Jacobsen EA,Ochkur SI,Willetts L,Shim K,Neely J,Kloeber J,Lesuer WE,Pero RS,Lacy P,Moqbel R,Lee NA,Lee JJ
17-5971 was used in Flow cytometry/Cell sorting to develop a novel murine strain to allow a better definition of the role of eosinophils in health and disease.
Mon Jul 01 00:00:00 UTC 2013
Ganciclovir prophylaxis improves late murine cytomegalovirus-induced renal allograft damage.
Transplantation
Shimamura M,Seleme MC,Guo L,Saunders U,Schoeb TR,George JF,Britt WJ
17-5971 was used in Flow cytometry/Cell sorting to examine the lasting effect of Ganciclovir treatment on renal allografts after the discontinuation of prophylaxis.
Tue Jan 15 00:00:00 UTC 2013
Antitumor activity of tumor-targeted RNA replicase-based plasmid that expresses interleukin-2 in a murine melanoma model.
Molecular pharmaceutics
Rodriguez BL,Blando JM,Lansakara-P DS,Kiguchi Y,DiGiovanni J,Cui Z
17-5971 was used in Flow cytometry/Cell sorting to evaluate the possible augmentation of pSIN-β function by incorporating the cytokine IL2.
Mon Jun 03 00:00:00 UTC 2013
G-CSF/anti-G-CSF antibody complexes drive the potent recovery and expansion of CD11b+Gr-1+ myeloid cells without compromising CD8+ T cell immune responses.
Journal of hematology & oncology
Rubinstein MP,Salem ML,Doedens AL,Moore CJ,Chiuzan C,Rivell GL,Cole DJ,Goldrath AW
17-5971 was used in Flow cytometry/Cell sorting to evaluate whether the biological activity of G-CSF can be improved by pre-association with anti-G-CSF monoclonal antibodies prior to injection.
Tue Oct 01 00:00:00 UTC 2013
Increased protection from vaccinia virus infection in mice genetically prone to lymphoproliferative disorders.
Journal of virology
Seedhom MO,Mathurin KS,Kim SK,Welsh RM
17-5971 was used in Flow cytometry/Cell sorting to reveal IFN-γ produced by Fas-deficient hosts enhances resistance to some viral infections.
Fri Jun 01 00:00:00 UTC 2012
Protective T cell immunity in mice following protein-TLR7/8 agonist-conjugate immunization requires aggregation, type I IFN, and multiple DC subsets.
The Journal of clinical investigation
Kastenmüller K,Wille-Reece U,Lindsay RW,Trager LR,Darrah PA,Flynn BJ,Becker MR,Udey MC,Clausen BE,Igyarto BZ,Kaplan DH,Kastenmüller W,Germain RN,Seder RA
17-5971 was used in Flow cytometry/Cell sorting to investigate the requirements for a vaccine to induce protective multifunctional T cell immunity in vivo.
Sun May 01 00:00:00 UTC 2011
Vesicular stomatitis virus expressing tumor suppressor p53 is a highly attenuated, potent oncolytic agent.
Journal of virology
Heiber JF,Barber GN
17-5971 was used in Flow cytometry/Cell sorting to investigate the potential of p53-incorporating-VSV for the design of VSV therapies and as a basis for VSV-based vaccines.
Sat Oct 01 00:00:00 UTC 2011
Impaired basophil induction leads to an age-dependent innate defect in type 2 immunity during helminth infection in mice.
Journal of immunology (Baltimore, Md. : 1950)
Nel HJ,Hams E,Saunders SP,Mangan NE,Smith P,Atzberger A,Flavell RA,Akira S,McKenzie AN,Fallon PG
17-5971 was used in Flow cytometry/Cell sorting to investigate the age-dependence of helminth-induced type 2 immune responses in adult and juvenile mice.
Fri Apr 15 00:00:00 UTC 2011
Genetic variation determines mast cell functions in experimental asthma.
Journal of immunology (Baltimore, Md. : 1950)
Becker M,Reuter S,Friedrich P,Doener F,Michel A,Bopp T,Klein M,Schmitt E,Schild H,Radsak MP,Echtenacher B,Taube C,Stassen M
17-5971 was used in Flow cytometry/Cell sorting to investigate the role of mast cells in allergic asthma.
Wed Jun 15 00:00:00 UTC 2011
Topical imiquimod has therapeutic and immunomodulatory effects against intracranial tumors.
Journal of immunotherapy (Hagerstown, Md. : 1997)
Xiong Z,Ohlfest JR
17-5971 was used in Flow cytometry/Cell sorting to demonstrate that topical application of the TLR7 agonist imiquimod can enhance T-cell responses to intracranial tumors as a single agent in mice.
Fri Apr 01 00:00:00 UTC 2011
Complexity of transcriptional regulation within the Rag locus: identification of a second Nwc promoter region within the Rag2 intron.
Immunogenetics
Laszkiewicz A,Cebrat M,Miazek A,Kisielow P
17-5971 was used in Flow cytometry/Cell sorting to study the transcriptional regulation of the Nwc gene in the Rag locus using the Nwc(tmpro1) mouse.
Tue Mar 01 00:00:00 UTC 2011
Innate immune activation by the viral PAMP poly I:C potentiates pulmonary graft-versus-host disease after allogeneic hematopoietic cell transplant.
Transplant immunology
Kinnier CV,Martinu T,Gowdy KM,Nugent JL,Kelly FL,Palmer SM
17-5971 was used in Flow cytometry/Cell sorting to suggest a novel pathway in which the innate antiviral response to a single viral PAMP can potentiate pulmonary GVHD after allogeneic HCT.
Sat Jan 15 00:00:00 UTC 2011
Study of camelpox virus pathogenesis in athymic nude mice.
PloS one
Duraffour S,Matthys P,van den Oord JJ,De Schutter T,Mitera T,Snoeck R,Andrei G
17-5971 was used in Flow cytometry/Cell sorting to confirm that NMRI immunocompetent mice are resistant to intranasal (i.n.) CMLV infection.
Thu Dec 01 00:00:00 UTC 2011
The death effector domain protein PEA-15 negatively regulates T-cell receptor signaling.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Pastorino S,Renganathan H,Caliva MJ,Filbert EL,Opoku-Ansah J,Sulzmaier FJ,Gawecka JE,Werlen G,Shaw AS,Ramos JW
17-5971 was used in Flow cytometry/Cell sorting to investigate the physiological function of PEA-15, showing that it negatively regulates T-cell receptor signalling.
Sun Aug 01 00:00:00 UTC 2010
Mast cell-derived TNF can exacerbate mortality during severe bacterial infections in C57BL/6-KitW-sh/W-sh mice.
The American journal of pathology
Piliponsky AM,Chen CC,Grimbaldeston MA,Burns-Guydish SM,Hardy J,Kalesnikoff J,Contag CH,Tsai M,Galli SJ
17-5971 was used in Flow cytometry/Cell sorting to investigate the roles of mast cells and their derived proteins in two models of severe bacterial infection.
Mon Feb 01 00:00:00 UTC 2010
Antigen-specific immunity and cross-priming by epithelial ovarian carcinoma-induced CD11b(+)Gr-1(+) cells.
Journal of immunology (Baltimore, Md. : 1950)
Tomihara K,Guo M,Shin T,Sun X,Ludwig SM,Brumlik MJ,Zhang B,Curiel TJ,Shin T
17-5971 was used in Flow cytometry/Cell sorting to investigate CD11b(+)Gr-1(+) cells found in epithelial ovarian cancer-bearing mice, which are induced by ovarian cancer cells and undergo antigen-specific immunity and cross-priming.
Tue Jun 01 00:00:00 UTC 2010
Chronic cigarette smoke exposure primes NK cell activation in a mouse model of chronic obstructive pulmonary disease.
Journal of immunology (Baltimore, Md. : 1950)
Motz GT,Eppert BL,Wortham BW,Amos-Kroohs RM,Flury JL,Wesselkamper SC,Borchers MT
17-5971 was used in Flow cytometry/Cell sorting to demonstrate an entirely novel mechanism of NK cell activation resulting from chronic cigarette smoke exposure.
Thu Apr 15 00:00:00 UTC 2010
Intracellular staining for analysis of the expression and phosphorylation of signal transducers and activators of transcription (STATs) in NK cells.
Methods in molecular biology (Clifton, N.J.)
Miyagi T,Lee SH,Biron CA
17-5971 was used in Flow cytometry/Cell sorting to develop approaches to measure the levels of expression and activation of signalling molecules.
Fri Feb 26 00:00:00 UTC 2010
Dendritic cells support the in vivo development and maintenance of NK cells via IL-15 trans-presentation.
Journal of immunology (Baltimore, Md. : 1950)
Castillo EF,Stonier SW,Frasca L,Schluns KS
17-5971 was used in Flow cytometry/Cell sorting to identify key role of DCs for trans-presentation and generation of NK cells.
Thu Oct 15 00:00:00 UTC 2009
Nylon wool purification alters the activation of T cells.
Molecular immunology
Wohler JE,Barnum SR
17-5971 was used in Flow cytometry/Cell sorting to examine the effects of nylon wool purification on T cell proliferation, activation marker expression and cytokine production.
Sun Feb 01 00:00:00 UTC 2009
Some putative prebiotics increase the severity of Salmonella enterica serovar Typhimurium infection in mice.
BMC microbiology
Petersen A,Heegaard PM,Pedersen AL,Andersen JB,Sørensen RB,Frøkiaer H,Lahtinen SJ,Ouwehand AC,Poulsen M,Licht TR
17-5971 was used in Flow cytometry/Cell sorting to examine whether Salmonella infection could be prevented by administration of dietary carbohydrates with different structures and digestibility profiles.
Mon Nov 30 00:00:00 UTC 2009
IL-7 and IL-15 differentially regulate CD8+ T-cell subsets during contraction of the immune response.
Blood
Rubinstein MP,Lind NA,Purton JF,Filippou P,Best JA,McGhee PA,Surh CD,Goldrath AW
17-5971 was used in Flow cytometry/Cell sorting to study IL-7 and IL-15 regulation of CD8+ T-cell subsets during contraction of the immune response.
Sat Nov 01 00:00:00 UTC 2008
Evidence for differential roles for NKG2D receptor signaling in innate host defense against coronavirus-induced neurological and liver disease.
Journal of virology
Walsh KB,Lodoen MB,Edwards RA,Lanier LL,Lane TE
17-5971 was used in Flow cytometry/Cell sorting to investigate the underlying mechanisms associated with protection from disease in MHV-CXCL10-infected mice.
Sat Mar 01 00:00:00 UTC 2008
Ly49 cluster sequence analysis in a mouse model of diabetes: an expanded repertoire of activating receptors in the NOD genome.
Genes and immunity
Belanger S,Tai LH,Anderson SK,Makrigiannis AP
17-5971 was used in Flow cytometry/Cell sorting to assess the contribution of the natural killer cell receptor Ly49 to the acceleration of disease in non-obese diabetic mice.
Mon Sep 01 00:00:00 UTC 2008
Converting IL-15 to a superagonist by binding to soluble IL-15R{alpha}.
Proceedings of the National Academy of Sciences of the United States of America
Rubinstein MP,Kovar M,Purton JF,Cho JH,Boyman O,Surh CD,Sprent J
17-5971 was used in Flow cytometry/Cell sorting to study how IL-2 function is markedly inhibited by binding to soluble IL-2Ralpha.
Tue Jun 13 00:00:00 UTC 2006