Product References
Generation of human tonsil epithelial organoids as an ex vivo model for SARS-CoV-2 infection.
Biomaterials
Kim HK,Kim H,Lee MK,Choi WH,Jang Y,Shin JS,Park JY,Bae DH,Hyun SI,Kim KH,Han HW,Lim B,Choi G,Kim M,Chang Lim Y,Yoo J
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Immunohistochemistry
Fri Apr 01 00:00:00 EDT 2022
Isolation and characterization mesenchymal stem cells from red panda (Ailurus fulgens styani) endometrium.
Conservation physiology
Wang DH,Wu XM,Chen JS,Cai ZG,An JH,Zhang MY,Li Y,Li FP,Hou R,Liu YL
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Sat Jan 01 00:00:00 EST 2022
Tumor suppressor DEAR1 regulates mammary epithelial cell fate and predicts early onset and metastasis in triple negative breast cancer.
Scientific reports
Le UQ,Chen N,Balasenthil S,Lurie E,Yang F,Liu S,Rubin L,Solis Soto LM,Raso MG,Batra H,Sahin AA,Wistuba II,Killary AM
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Mon Nov 14 00:00:00 EST 2022
Ablating Lgr5-expressing prostatic stromal cells activates the ERK-mediated mechanosensory signaling and disrupts prostate tissue homeostasis.
Cell reports
Wei X,Zhang L,Zhang Y,Cooper C,Brewer C,Tsai CF,Wang YT,Glaz M,Wessells HB,Que J,Titus MA,Cirulli V,Glaser A,Liu T,Reder NP,Creighton CJ,Xin L
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Tue Sep 06 00:00:00 EDT 2022
The Effect of Infant Gastric Digestion on Human Maternal Milk Cells.
Molecular nutrition & food research
Doerfler R,Melamed JR,Whitehead KA
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Sat Oct 01 00:00:00 EDT 2022
Embigin is a fibronectin receptor that affects sebaceous gland differentiation and metabolism.
Developmental cell
Sipilä K,Rognoni E,Jokinen J,Tewary M,Vietri Rudan M,Talvi S,Jokinen V,Dahlström KM,Liakath-Ali K,Mobasseri A,Du-Harpur X,Käpylä J,Nutt SL,Salminen TA,Heino J,Watt FM
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Immunohistochemistry
Mon Jun 20 00:00:00 EDT 2022
ROR2 regulates self-renewal and maintenance of hair follicle stem cells.
Nature communications
Veltri A,Lang CMR,Cangiotti G,Chan CK,Lien WH
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2022
Endogenous retroviruses promote homeostatic and inflammatory responses to the microbiota.
Cell
Lima-Junior DS,Krishnamurthy SR,Bouladoux N,Collins N,Han SJ,Chen EY,Constantinides MG,Link VM,Lim AI,Enamorado M,Cataisson C,Gil L,Rao I,Farley TK,Koroleva G,Attig J,Yuspa SH,Fischbach MA,Kassiotis G,Belkaid Y
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Thu Jul 08 00:00:00 EDT 2021
CBP-mediated Wnt3a/β-catenin signaling promotes cervical oncogenesis initiated by Piwil2.
Neoplasia (New York, N.Y.)
Feng D,Yan K,Liang H,Liang J,Wang W,Yu H,Zhou Y,Zhao W,Dong Z,Ling B
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
GRHL3 activates FSCN1 to relax cell-cell adhesions between migrating keratinocytes during wound reepithelialization.
JCI insight
Kashgari G,Venkatesh S,Refuerzo S,Pham B,Bayat A,Klein RH,Ramos R,Ta AP,Plikus MV,Wang PH,Andersen B
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Wed Sep 08 00:00:00 EDT 2021
Generation and functional characterization of murine mammary organoids.
STAR protocols
Yip HYK,Papa A
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Immunocytochemistry
Fri Sep 17 00:00:00 EDT 2021
Single-cell analyses unravel cell type-specific responses to a vitamin D analog in prostatic precancerous lesions.
Science advances
Abu El Maaty MA,Grelet E,Keime C,Rerra AI,Gantzer J,Emprou C,Terzic J,Lutzing R,Bornert JM,Laverny G,Metzger D
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2021
Nov/CCN3 Enhances Cord Blood Engraftment by Rapidly Recruiting Latent Human Stem Cell Activity.
Cell stem cell
Gupta R,Turati V,Brian D,Thrussel C,Wilbourn B,May G,Enver T
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Neutralization
Thu Apr 02 00:00:00 EDT 2020
Induced Pluripotent Stem Cells Derived From Two Idiopathic Azoospermia Patients Display Compromised Differentiation Potential for Primordial Germ Cell Fate.
Frontiers in cell and developmental biology
Fang F,Li Z,Zhao Q,Ye Z,Gu X,Pan F,Li H,Xiang W,Xiong C
11-0495 was used in Flow cytometry/Cell sorting to evaluate the differentiation potential for human germline of idiopathic non-obstructive azoospermia (NOA) patient-specific induced pluripotent stem cells (iPSCs).
Mon Sep 28 00:00:00 EDT 2020
An IFT20 mechanotrafficking axis is required for integrin recycling, focal adhesion dynamics, and polarized cell migration.
Molecular biology of the cell
Su S,Begum S,Ezratty EJ
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2020
An IFT20 mechanotrafficking axis is required for integrin recycling, focal adhesion dynamics, and polarized cell migration.
Molecular biology of the cell
Su S,Begum S,Ezratty EJ
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2020
Chronic expression of p16INK4a in the epidermis induces Wnt-mediated hyperplasia and promotes tumor initiation.
Nature communications
Azazmeh N,Assouline B,Winter E,Ruppo S,Nevo Y,Maly A,Meir K,Witkiewicz AK,Cohen J,Rizou SV,Pikarsky E,Luxenburg C,Gorgoulis VG,Ben-Porath I
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2020
Chemotherapeutic Stress Influences Epithelial-Mesenchymal Transition and Stemness in Cancer Stem Cells of Triple-Negative Breast Cancer.
International journal of molecular sciences
Li X,Strietz J,Bleilevens A,Stickeler E,Maurer J
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Wed Jan 08 00:00:00 EST 2020
Polyisocyanide Hydrogels as a Tunable Platform for Mammary Gland Organoid Formation.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Tang C,Span PN,Rowan AE,Aalders TW,Schalken JA,Adema GJ,Kouwer PHJ,Zegers MMP,Ansems M
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2020
Discrete populations of isotype-switched memory B lymphocytes are maintained in murine spleen and bone marrow.
Nature communications
Riedel R,Addo R,Ferreira-Gomes M,Heinz GA,Heinrich F,Kummer J,Greiff V,Schulz D,Klaeden C,Cornelis R,Menzel U,Kröger S,Stervbo U,Köhler R,Haftmann C,Kühnel S,Lehmann K,Maschmeyer P,McGrath M,Naundorf S,Hahne S,Sercan-Alp Ö,Siracusa F,Stefanowski J,Weber M,Westendorf K,Zimmermann J,Hauser AE,Reddy ST,Durek P,Chang HD,Mashreghi MF,Radbruch A
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Fri May 22 00:00:00 EDT 2020
Evolutionarily conserved sequence motif analysis guides development of chemically defined hydrogels for therapeutic vascularization.
Science advances
Jia J,Jeon EJ,Li M,Richards DJ,Lee S,Jung Y,Barrs RW,Coyle R,Li X,Chou JC,Yost MJ,Gerecht S,Cho SW,Mei Y
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Neutralization
Wed Jul 01 00:00:00 EDT 2020
Comprehensive Cell Surface Antigen Analysis Identifies Transferrin Receptor Protein-1 (CD71) as a Negative Selection Marker for Human Neuronal Cells.
Stem cells (Dayton, Ohio)
Menon V,Thomas R,Elgueta C,Horl M,Osborn T,Hallett PJ,Bartos M,Isacson O,Pruszak J
Tue Oct 01 00:00:00 EDT 2019
The MMTV-Wnt1 murine model produces two phenotypically distinct subtypes of mammary tumors with unique therapeutic responses to an EGFR inhibitor.
Disease models & mechanisms
Pfefferle AD,Darr DB,Calhoun BC,Mott KR,Rosen JM,Perou CM
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Fri Jul 05 00:00:00 EDT 2019
A basal-enriched microRNA is required for prostate tumorigenesis in a Pten knockout mouse model.
Molecular carcinogenesis
Fan X,Bjerke GA,Riemondy K,Wang L,Yi R
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Sun Dec 01 00:00:00 EST 2019
Regulatory T Cells Control the Switch From in situ to Invasive Breast Cancer.
Frontiers in immunology
Martinez LM,Robila V,Clark NM,Du W,Idowu MO,Rutkowski MR,Bos PD
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2020
Ubiquitin ligase RNF8 suppresses Notch signaling to regulate mammary development and tumorigenesis.
The Journal of clinical investigation
Li L,Guturi KKN,Gautreau B,Patel PS,Saad A,Morii M,Mateo F,Palomero L,Barbour H,Gomez A,Ng D,Kotlyar M,Pastrello C,Jackson HW,Khokha R,Jurisica I,Affar EB,Raught B,Sanchez O,Alaoui-Jamali M,Pujana MA,Hakem A,Hakem R
11-0495 was used in Flow cytometry/Cell sorting to study the correlation between low expression of full-length RNF8 and poor prognosis for breast cancer patients.
Mon Oct 01 00:00:00 EDT 2018
Integrin-Rac signalling for mammary epithelial stem cell self-renewal.
Breast cancer research : BCR
Olabi S,Ucar A,Brennan K,Streuli CH
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Mon Oct 22 00:00:00 EDT 2018
The hypoxic tumor microenvironment in vivo selects the cancer stem cell fate of breast cancer cells.
Breast cancer research : BCR
Kim H,Lin Q,Glazer PM,Yun Z
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Tue Mar 06 00:00:00 EST 2018
Profiling human breast epithelial cells using single cell RNA sequencing identifies cell diversity.
Nature communications
Nguyen QH,Pervolarakis N,Blake K,Ma D,Davis RT,James N,Phung AT,Willey E,Kumar R,Jabart E,Driver I,Rock J,Goga A,Khan SA,Lawson DA,Werb Z,Kessenbrock K
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Wed May 23 00:00:00 EDT 2018
Single-Cell Analysis Identifies LY6D as a Marker Linking Castration-Resistant Prostate Luminal Cells to Prostate Progenitors and Cancer.
Cell reports
Barros-Silva JD,Linn DE,Steiner I,Guo G,Ali A,Pakula H,Ashton G,Peset I,Brown M,Clarke NW,Bronson RT,Yuan GC,Orkin SH,Li Z,Baena E
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Tue Dec 18 00:00:00 EST 2018
Inflammatory memory sensitizes skin epithelial stem cells to tissue damage.
Nature
Naik S,Larsen SB,Gomez NC,Alaverdyan K,Sendoel A,Yuan S,Polak L,Kulukian A,Chai S,Fuchs E
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Thu Oct 26 00:00:00 EDT 2017
Regulatory T Cells in Skin Facilitate Epithelial Stem Cell Differentiation.
Cell
Ali N,Zirak B,Rodriguez RS,Pauli ML,Truong HA,Lai K,Ahn R,Corbin K,Lowe MM,Scharschmidt TC,Taravati K,Tan MR,Ricardo-Gonzalez RR,Nosbaum A,Bertolini M,Liao W,Nestle FO,Paus R,Cotsarelis G,Abbas AK,Rosenblum MD
11-0495 was used in Flow cytometry/Cell sorting to study the role of regulatory T cells in hair follicles and hair follicle stem cell biology.
Thu Jun 01 00:00:00 EDT 2017
Autoreactive T Cells and Chronic Fungal Infection Drive Esophageal Carcinogenesis.
Cell host & microbe
Zhu F,Willette-Brown J,Song NY,Lomada D,Song Y,Xue L,Gray Z,Zhao Z,Davis SR,Sun Z,Zhang P,Wu X,Zhan Q,Richie ER,Hu Y
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Wed Apr 12 00:00:00 EDT 2017
Hair follicle stem cell cultures reveal self-organizing plasticity of stem cells and their progeny.
The EMBO journal
Chacón-Martínez CA,Klose M,Niemann C,Glauche I,Wickström SA
Tue Jan 17 00:00:00 EST 2017
Low Testosterone Alters the Activity of Mouse Prostate Stem Cells.
The Prostate
Zhou Y,Copeland B,Otto-Duessel M,He M,Markel S,Synold TW,Jones JO
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
Induced p53 loss in mouse luminal cells causes clonal expansion and development of mammary tumours.
Nature communications
Tao L,Xiang D,Xie Y,Bronson RT,Li Z
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Mon Feb 13 00:00:00 EST 2017
Defining stem cell dynamics and migration during wound healing in mouse skin epidermis.
Nature communications
Aragona M,Dekoninck S,Rulands S,Lenglez S,Mascré G,Simons BD,Blanpain C
Wed Mar 01 00:00:00 EST 2017
Characterization of the CD49f+/CD44+/CD24- single-cell derived stem cell population in basal-like DCIS cells.
Oncotarget
Duru N,Gernapudi R,Lo PK,Yao Y,Wolfson B,Zhang Y,Zhou Q
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Tue Jul 26 00:00:00 EDT 2016
Inhibition of the glucocorticoid receptor results in an enhanced miR-99a/100-mediated radiation response in stem-like cells from human prostate cancers.
Oncotarget
Rane JK,Erb HH,Nappo G,Mann VM,Simms MS,Collins AT,Visakorpi T,Maitland NJ
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
Directed elimination of senescent cells by inhibition of BCL-W and BCL-XL.
Nature communications
Yosef R,Pilpel N,Tokarsky-Amiel R,Biran A,Ovadya Y,Cohen S,Vadai E,Dassa L,Shahar E,Condiotti R,Ben-Porath I,Krizhanovsky V
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Wed Apr 06 00:00:00 EDT 2016
Yap1 promotes the survival and self-renewal of breast tumor initiating cells via inhibiting Smad3 signaling.
Oncotarget
Sun JG,Chen XW,Zhang LP,Wang J,Diehn M
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2016
Llgl1 prevents metaplastic survival driven by epidermal growth factor dependent migration.
Oncotarget
Greenwood E,Maisel S,Ebertz D,Russ A,Pandey R,Schroeder J
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Tue Sep 20 00:00:00 EDT 2016
The Transcriptional Repressor ZNF503/Zeppo2 Promotes Mammary Epithelial Cell Proliferation and Enhances Cell Invasion.
The Journal of biological chemistry
Shahi P,Slorach EM,Wang CY,Chou J,Lu A,Ruderisch A,Werb Z
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Fri Feb 06 00:00:00 EST 2015
Merlin Isoforms 1 and 2 Both Act as Tumour Suppressors and Are Required for Optimal Sperm Maturation.
PloS one
Zoch A,Mayerl S,Schulz A,Greither T,Frappart L,Rübsam J,Heuer H,Giovannini M,Morrison H
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Mon May 23 00:00:00 EDT 2016
SF3B1 mutant MDS-initiating cells may arise from the haematopoietic stem cell compartment.
Nature communications
Mian SA,Rouault-Pierre K,Smith AE,Seidl T,Pizzitola I,Kizilors A,Kulasekararaj AG,Bonnet D,Mufti GJ
11-0495 was used in Flow cytometry/Cell sorting to demonstrate that mutations to SF3B1 can arise from CD34(+)CD38(-)CD45RA(-)CD90(+)CD49f(+) haematopoietic stem cells and is an initiating event in myelodysplastic syndrome patients.
Tue Dec 08 00:00:00 EST 2015
NDY1/KDM2B functions as a master regulator of polycomb complexes and controls self-renewal of breast cancer stem cells.
Cancer research
Kottakis F,Foltopoulou P,Sanidas I,Keller P,Wronski A,Dake BT,Ezell SA,Shen Z,Naber SP,Hinds PW,McNiel E,Kuperwasser C,Tsichlis PN
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Tue Jul 15 00:00:00 EDT 2014
FGF ligands of the postnatal mammary stroma regulate distinct aspects of epithelial morphogenesis.
Development (Cambridge, England)
Zhang X,Martinez D,Koledova Z,Qiao G,Streuli CH,Lu P
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Fri Feb 01 00:00:00 EST 2013
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Fri Feb 01 00:00:00 EST 2013
Evidence for a multipotent mammary progenitor with pregnancy-specific activity.
Breast cancer research : BCR
Kaanta AS,Virtanen C,Selfors LM,Brugge JS,Neel BG
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Mon Dec 08 00:00:00 EST 2014
Evidence for a multipotent mammary progenitor with pregnancy-specific activity.
Breast cancer research : BCR
Kaanta AS,Virtanen C,Selfors LM,Brugge JS,Neel BG
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Mon Dec 08 00:00:00 EST 2014
Generation of a novel germline stem cell line expressing a germline-specific reporter in the mouse.
Genesis (New York, N.Y. : 2000)
Shiura H,Ikeda R,Lee J,Sato T,Ogonuki N,Hirose M,Ogura A,Ogawa T,Abe K
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2013
Distinct effects of EGFR ligands on human mammary epithelial cell differentiation.
PloS one
Mukhopadhyay C,Zhao X,Maroni D,Band V,Naramura M
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Fri Jul 11 00:00:00 EDT 2014
Highly upregulated Lhx2 in the Foxn1-/- nude mouse phenotype reflects a dysregulated and expanded epidermal stem cell niche.
PloS one
Bohr S,Patel SJ,Vasko R,Shen K,Huang G,Yarmush ML,Berthiaume F
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Mon Dec 23 00:00:00 EST 2013
Stem-like epithelial cells are concentrated in the distal end of the fallopian tube: a site for injury and serous cancer initiation.
Stem cells (Dayton, Ohio)
Paik DY,Janzen DM,Schafenacker AM,Velasco VS,Shung MS,Cheng D,Huang J,Witte ON,Memarzadeh S
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 11-0495-82) in Flow Cytometry
Thu Nov 01 00:00:00 EDT 2012