Product References
Integrin expression and extracellular matrix adhesion of septoclasts, pericytes, and endothelial cells at the chondro-osseous junction and the metaphysis of the proximal tibia in young mice.
Journal of anatomy
Bando Y,Nagasaka A,Onozawa G,Sakiyama K,Owada Y,Amano O
14-0495-82 was used in Immunohistochemistry to suggest that the adhesion between septoclasts and the cartilage ECM forming the scaffolds for cartilage resorption and migration is provided by integrin α2-collagen-II/-X interaction and that the adhesions between the BM and pericytes or endothelial cells are mediated by integrin α1-collagen-IV and integrin α2/α6-laminin interaction, respectively.
Mon May 01 00:00:00 EDT 2023
Generation of human tonsil epithelial organoids as an ex vivo model for SARS-CoV-2 infection.
Biomaterials
Kim HK,Kim H,Lee MK,Choi WH,Jang Y,Shin JS,Park JY,Bae DH,Hyun SI,Kim KH,Han HW,Lim B,Choi G,Kim M,Chang Lim Y,Yoo J
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Immunohistochemistry
Fri Apr 01 00:00:00 EDT 2022
Isolation and characterization mesenchymal stem cells from red panda (Ailurus fulgens styani) endometrium.
Conservation physiology
Wang DH,Wu XM,Chen JS,Cai ZG,An JH,Zhang MY,Li Y,Li FP,Hou R,Liu YL
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Sat Jan 01 00:00:00 EST 2022
Tumor suppressor DEAR1 regulates mammary epithelial cell fate and predicts early onset and metastasis in triple negative breast cancer.
Scientific reports
Le UQ,Chen N,Balasenthil S,Lurie E,Yang F,Liu S,Rubin L,Solis Soto LM,Raso MG,Batra H,Sahin AA,Wistuba II,Killary AM
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Mon Nov 14 00:00:00 EST 2022
Ablating Lgr5-expressing prostatic stromal cells activates the ERK-mediated mechanosensory signaling and disrupts prostate tissue homeostasis.
Cell reports
Wei X,Zhang L,Zhang Y,Cooper C,Brewer C,Tsai CF,Wang YT,Glaz M,Wessells HB,Que J,Titus MA,Cirulli V,Glaser A,Liu T,Reder NP,Creighton CJ,Xin L
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Tue Sep 06 00:00:00 EDT 2022
The Effect of Infant Gastric Digestion on Human Maternal Milk Cells.
Molecular nutrition & food research
Doerfler R,Melamed JR,Whitehead KA
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Sat Oct 01 00:00:00 EDT 2022
Embigin is a fibronectin receptor that affects sebaceous gland differentiation and metabolism.
Developmental cell
Sipilä K,Rognoni E,Jokinen J,Tewary M,Vietri Rudan M,Talvi S,Jokinen V,Dahlström KM,Liakath-Ali K,Mobasseri A,Du-Harpur X,Käpylä J,Nutt SL,Salminen TA,Heino J,Watt FM
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Immunohistochemistry
Mon Jun 20 00:00:00 EDT 2022
ROR2 regulates self-renewal and maintenance of hair follicle stem cells.
Nature communications
Veltri A,Lang CMR,Cangiotti G,Chan CK,Lien WH
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2022
Endogenous retroviruses promote homeostatic and inflammatory responses to the microbiota.
Cell
Lima-Junior DS,Krishnamurthy SR,Bouladoux N,Collins N,Han SJ,Chen EY,Constantinides MG,Link VM,Lim AI,Enamorado M,Cataisson C,Gil L,Rao I,Farley TK,Koroleva G,Attig J,Yuspa SH,Fischbach MA,Kassiotis G,Belkaid Y
14-0495-82 was used in Flow Cytometry to determine response to chemical signalling caused by endogenous retrovirus in skin microbiota.
Thu Jul 08 00:00:00 EDT 2021
Endogenous retroviruses promote homeostatic and inflammatory responses to the microbiota.
Cell
Lima-Junior DS,Krishnamurthy SR,Bouladoux N,Collins N,Han SJ,Chen EY,Constantinides MG,Link VM,Lim AI,Enamorado M,Cataisson C,Gil L,Rao I,Farley TK,Koroleva G,Attig J,Yuspa SH,Fischbach MA,Kassiotis G,Belkaid Y
14-0495-82 was used in Flow Cytometry to determine response to chemical signalling caused by endogenous retrovirus in skin microbiota.
Thu Jul 08 00:00:00 EDT 2021
CBP-mediated Wnt3a/β-catenin signaling promotes cervical oncogenesis initiated by Piwil2.
Neoplasia (New York, N.Y.)
Feng D,Yan K,Liang H,Liang J,Wang W,Yu H,Zhou Y,Zhao W,Dong Z,Ling B
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
GRHL3 activates FSCN1 to relax cell-cell adhesions between migrating keratinocytes during wound reepithelialization.
JCI insight
Kashgari G,Venkatesh S,Refuerzo S,Pham B,Bayat A,Klein RH,Ramos R,Ta AP,Plikus MV,Wang PH,Andersen B
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Wed Sep 08 00:00:00 EDT 2021
Generation and functional characterization of murine mammary organoids.
STAR protocols
Yip HYK,Papa A
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Immunocytochemistry
Fri Sep 17 00:00:00 EDT 2021
Single-cell analyses unravel cell type-specific responses to a vitamin D analog in prostatic precancerous lesions.
Science advances
Abu El Maaty MA,Grelet E,Keime C,Rerra AI,Gantzer J,Emprou C,Terzic J,Lutzing R,Bornert JM,Laverny G,Metzger D
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2021
Nov/CCN3 Enhances Cord Blood Engraftment by Rapidly Recruiting Latent Human Stem Cell Activity.
Cell stem cell
Gupta R,Turati V,Brian D,Thrussel C,Wilbourn B,May G,Enver T
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Neutralization
Thu Apr 02 00:00:00 EDT 2020
An IFT20 mechanotrafficking axis is required for integrin recycling, focal adhesion dynamics, and polarized cell migration.
Molecular biology of the cell
Su S,Begum S,Ezratty EJ
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2020
Chronic expression of p16INK4a in the epidermis induces Wnt-mediated hyperplasia and promotes tumor initiation.
Nature communications
Azazmeh N,Assouline B,Winter E,Ruppo S,Nevo Y,Maly A,Meir K,Witkiewicz AK,Cohen J,Rizou SV,Pikarsky E,Luxenburg C,Gorgoulis VG,Ben-Porath I
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2020
Chemotherapeutic Stress Influences Epithelial-Mesenchymal Transition and Stemness in Cancer Stem Cells of Triple-Negative Breast Cancer.
International journal of molecular sciences
Li X,Strietz J,Bleilevens A,Stickeler E,Maurer J
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Wed Jan 08 00:00:00 EST 2020
Polyisocyanide Hydrogels as a Tunable Platform for Mammary Gland Organoid Formation.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Tang C,Span PN,Rowan AE,Aalders TW,Schalken JA,Adema GJ,Kouwer PHJ,Zegers MMP,Ansems M
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2020
Discrete populations of isotype-switched memory B lymphocytes are maintained in murine spleen and bone marrow.
Nature communications
Riedel R,Addo R,Ferreira-Gomes M,Heinz GA,Heinrich F,Kummer J,Greiff V,Schulz D,Klaeden C,Cornelis R,Menzel U,Kröger S,Stervbo U,Köhler R,Haftmann C,Kühnel S,Lehmann K,Maschmeyer P,McGrath M,Naundorf S,Hahne S,Sercan-Alp Ö,Siracusa F,Stefanowski J,Weber M,Westendorf K,Zimmermann J,Hauser AE,Reddy ST,Durek P,Chang HD,Mashreghi MF,Radbruch A
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Fri May 22 00:00:00 EDT 2020
Contribution of GATA6 to homeostasis of the human upper pilosebaceous unit and acne pathogenesis.
Nature communications
Oulès B,Philippeos C,Segal J,Tihy M,Vietri Rudan M,Cujba AM,Grange PA,Quist S,Natsuga K,Deschamps L,Dupin N,Donati G,Watt FM
14-0495-81 was used in Immunohistochemistry (PFA fixed) to show that GATA6, which is expressed in the upper pilosebaceous unit of normal human skin, is down-regulated in acne.
Tue Oct 20 00:00:00 EDT 2020
Evolutionarily conserved sequence motif analysis guides development of chemically defined hydrogels for therapeutic vascularization.
Science advances
Jia J,Jeon EJ,Li M,Richards DJ,Lee S,Jung Y,Barrs RW,Coyle R,Li X,Chou JC,Yost MJ,Gerecht S,Cho SW,Mei Y
14-0495 was used in Inhibition assays to identify a peptide (a1) that showed superior supports for endothelial cell (EC) functions.
Wed Jul 01 00:00:00 EDT 2020
Evolutionarily conserved sequence motif analysis guides development of chemically defined hydrogels for therapeutic vascularization.
Science advances
Jia J,Jeon EJ,Li M,Richards DJ,Lee S,Jung Y,Barrs RW,Coyle R,Li X,Chou JC,Yost MJ,Gerecht S,Cho SW,Mei Y
14-0495 was used in Inhibition assays to identify a peptide (a1) that showed superior supports for endothelial cell (EC) functions.
Wed Jul 01 00:00:00 EDT 2020
Adaptive Immune Resistance Emerges from Tumor-Initiating Stem Cells.
Cell
Miao Y,Yang H,Levorse J,Yuan S,Polak L,Sribour M,Singh B,Rosenblum MD,Fuchs E
14-0495 was used in Flow cytometry/Cell sorting to place tSCs at the crux of how immune checkpoint pathways are activated.
Thu May 16 00:00:00 EDT 2019
Mutant Lef1 controls Gata6 in sebaceous gland development and cancer.
The EMBO journal
Oulès B,Rognoni E,Hoste E,Goss G,Fiehler R,Natsuga K,Quist S,Mentink R,Donati G,Watt FM
14-0495-82 was used in Flow Cytometry to show that Gata6 is a β-catenin-independent transcriptional target of mutant Lef1.
Thu May 02 00:00:00 EDT 2019
Mutant Lef1 controls Gata6 in sebaceous gland development and cancer.
The EMBO journal
Oulès B,Rognoni E,Hoste E,Goss G,Fiehler R,Natsuga K,Quist S,Mentink R,Donati G,Watt FM
14-0495-82 was used in Flow Cytometry to show that Gata6 is a β-catenin-independent transcriptional target of mutant Lef1.
Thu May 02 00:00:00 EDT 2019
The MMTV-Wnt1 murine model produces two phenotypically distinct subtypes of mammary tumors with unique therapeutic responses to an EGFR inhibitor.
Disease models & mechanisms
Pfefferle AD,Darr DB,Calhoun BC,Mott KR,Rosen JM,Perou CM
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Fri Jul 05 00:00:00 EDT 2019
A basal-enriched microRNA is required for prostate tumorigenesis in a Pten knockout mouse model.
Molecular carcinogenesis
Fan X,Bjerke GA,Riemondy K,Wang L,Yi R
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Sun Dec 01 00:00:00 EST 2019
Regulatory T Cells Control the Switch From in situ to Invasive Breast Cancer.
Frontiers in immunology
Martinez LM,Robila V,Clark NM,Du W,Idowu MO,Rutkowski MR,Bos PD
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2020
Integrin-Rac signalling for mammary epithelial stem cell self-renewal.
Breast cancer research : BCR
Olabi S,Ucar A,Brennan K,Streuli CH
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Mon Oct 22 00:00:00 EDT 2018
The hypoxic tumor microenvironment in vivo selects the cancer stem cell fate of breast cancer cells.
Breast cancer research : BCR
Kim H,Lin Q,Glazer PM,Yun Z
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Tue Mar 06 00:00:00 EST 2018
Profiling human breast epithelial cells using single cell RNA sequencing identifies cell diversity.
Nature communications
Nguyen QH,Pervolarakis N,Blake K,Ma D,Davis RT,James N,Phung AT,Willey E,Kumar R,Jabart E,Driver I,Rock J,Goga A,Khan SA,Lawson DA,Werb Z,Kessenbrock K
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Wed May 23 00:00:00 EDT 2018
Single-Cell Analysis Identifies LY6D as a Marker Linking Castration-Resistant Prostate Luminal Cells to Prostate Progenitors and Cancer.
Cell reports
Barros-Silva JD,Linn DE,Steiner I,Guo G,Ali A,Pakula H,Ashton G,Peset I,Brown M,Clarke NW,Bronson RT,Yuan GC,Orkin SH,Li Z,Baena E
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Tue Dec 18 00:00:00 EST 2018
Inflammatory memory sensitizes skin epithelial stem cells to tissue damage.
Nature
Naik S,Larsen SB,Gomez NC,Alaverdyan K,Sendoel A,Yuan S,Polak L,Kulukian A,Chai S,Fuchs E
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Thu Oct 26 00:00:00 EDT 2017
Autoreactive T Cells and Chronic Fungal Infection Drive Esophageal Carcinogenesis.
Cell host & microbe
Zhu F,Willette-Brown J,Song NY,Lomada D,Song Y,Xue L,Gray Z,Zhao Z,Davis SR,Sun Z,Zhang P,Wu X,Zhan Q,Richie ER,Hu Y
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Wed Apr 12 00:00:00 EDT 2017
Low Testosterone Alters the Activity of Mouse Prostate Stem Cells.
The Prostate
Zhou Y,Copeland B,Otto-Duessel M,He M,Markel S,Synold TW,Jones JO
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
Induced p53 loss in mouse luminal cells causes clonal expansion and development of mammary tumours.
Nature communications
Tao L,Xiang D,Xie Y,Bronson RT,Li Z
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Mon Feb 13 00:00:00 EST 2017
Stem cell function and stress response are controlled by protein synthesis.
Nature
Blanco S,Bandiera R,Popis M,Hussain S,Lombard P,Aleksic J,Sajini A,Tanna H,Cortés-Garrido R,Gkatza N,Dietmann S,Frye M
14-0495 was used in Immunohistofluorescence to study how stem cells can revoke translation inhibition pathways to control their function and stress response..
Thu Jun 16 00:00:00 EDT 2016
Characterization of the CD49f+/CD44+/CD24- single-cell derived stem cell population in basal-like DCIS cells.
Oncotarget
Duru N,Gernapudi R,Lo PK,Yao Y,Wolfson B,Zhang Y,Zhou Q
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Tue Jul 26 00:00:00 EDT 2016
Inhibition of the glucocorticoid receptor results in an enhanced miR-99a/100-mediated radiation response in stem-like cells from human prostate cancers.
Oncotarget
Rane JK,Erb HH,Nappo G,Mann VM,Simms MS,Collins AT,Visakorpi T,Maitland NJ
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
Directed elimination of senescent cells by inhibition of BCL-W and BCL-XL.
Nature communications
Yosef R,Pilpel N,Tokarsky-Amiel R,Biran A,Ovadya Y,Cohen S,Vadai E,Dassa L,Shahar E,Condiotti R,Ben-Porath I,Krizhanovsky V
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Wed Apr 06 00:00:00 EDT 2016
Yap1 promotes the survival and self-renewal of breast tumor initiating cells via inhibiting Smad3 signaling.
Oncotarget
Sun JG,Chen XW,Zhang LP,Wang J,Diehn M
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2016
Llgl1 prevents metaplastic survival driven by epidermal growth factor dependent migration.
Oncotarget
Greenwood E,Maisel S,Ebertz D,Russ A,Pandey R,Schroeder J
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Tue Sep 20 00:00:00 EDT 2016
Hyperglycemia Induces Skin Barrier Dysfunctions with Impairment of Epidermal Integrity in Non-Wounded Skin of Type 1 Diabetic Mice.
PloS one
Okano J,Kojima H,Katagi M,Nakagawa T,Nakae Y,Terashima T,Kurakane T,Kubota M,Maegawa H,Udagawa J
14-0495 was used in Flow cytometry/Cell sorting to investigate whether hyperglycaemia induces skin barrier dysfunction through alterations of epidermal integrity.
Wed Jun 21 00:00:00 EDT 2017
The Transcriptional Repressor ZNF503/Zeppo2 Promotes Mammary Epithelial Cell Proliferation and Enhances Cell Invasion.
The Journal of biological chemistry
Shahi P,Slorach EM,Wang CY,Chou J,Lu A,Ruderisch A,Werb Z
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Fri Feb 06 00:00:00 EST 2015
Merlin Isoforms 1 and 2 Both Act as Tumour Suppressors and Are Required for Optimal Sperm Maturation.
PloS one
Zoch A,Mayerl S,Schulz A,Greither T,Frappart L,Rübsam J,Heuer H,Giovannini M,Morrison H
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Mon May 23 00:00:00 EDT 2016
Enhanced Expression of Integrin αvβ3 Induced by TGF-β Is Required for the Enhancing Effect of Fibroblast Growth Factor 1 (FGF1) in TGF-β-Induced Epithelial-Mesenchymal Transition (EMT) in Mammary Epithelial Cells.
PloS one
Mori S,Kodaira M,Ito A,Okazaki M,Kawaguchi N,Hamada Y,Takada Y,Matsuura N
14-0495 was used in Immunocytochemistry to study the role of integrin αvβ3 in TGF-β-induced epithelial-mesenchymal transition in mammary epithelial cells.
Tue May 24 00:00:00 EDT 2016
NDY1/KDM2B functions as a master regulator of polycomb complexes and controls self-renewal of breast cancer stem cells.
Cancer research
Kottakis F,Foltopoulou P,Sanidas I,Keller P,Wronski A,Dake BT,Ezell SA,Shen Z,Naber SP,Hinds PW,McNiel E,Kuperwasser C,Tsichlis PN
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Tue Jul 15 00:00:00 EDT 2014
Force engages vinculin and promotes tumor progression by enhancing PI3K activation of phosphatidylinositol (3,4,5)-triphosphate.
Cancer research
Rubashkin MG,Cassereau L,Bainer R,DuFort CC,Yui Y,Ou G,Paszek MJ,Davidson MW,Chen YY,Weaver VM
14-0495 was used in Immunofluorescence to determine that extracellular matrix stiffness could induce tumour progression by promoting the assembly of signalling scaffolds.
Mon Sep 01 00:00:00 EDT 2014
FGF ligands of the postnatal mammary stroma regulate distinct aspects of epithelial morphogenesis.
Development (Cambridge, England)
Zhang X,Martinez D,Koledova Z,Qiao G,Streuli CH,Lu P
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
Oligomerization-induced conformational change in the C-terminal region of Nel-like molecule 1 (NELL1) protein is necessary for the efficient mediation of murine MC3T3-E1 cell adhesion and spreading.
The Journal of biological chemistry
Nakamura Y,Hasebe A,Takahashi K,Iijima M,Yoshimoto N,Maturana AD,Ting K,Kuroda S,Niimi T
14-0495-82 was used in Inhibition experiments to define the structural requirements for NELL1-mediated cell adhesion.
Fri Apr 04 00:00:00 EDT 2014
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Fri Feb 01 00:00:00 EST 2013
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Fri Feb 01 00:00:00 EST 2013
Evidence for a multipotent mammary progenitor with pregnancy-specific activity.
Breast cancer research : BCR
Kaanta AS,Virtanen C,Selfors LM,Brugge JS,Neel BG
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Immunofluorescence
Mon Dec 08 00:00:00 EST 2014
Evidence for a multipotent mammary progenitor with pregnancy-specific activity.
Breast cancer research : BCR
Kaanta AS,Virtanen C,Selfors LM,Brugge JS,Neel BG
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Immunofluorescence
Mon Dec 08 00:00:00 EST 2014
Generation of a novel germline stem cell line expressing a germline-specific reporter in the mouse.
Genesis (New York, N.Y. : 2000)
Shiura H,Ikeda R,Lee J,Sato T,Ogonuki N,Hirose M,Ogura A,Ogawa T,Abe K
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2013
Distinct effects of EGFR ligands on human mammary epithelial cell differentiation.
PloS one
Mukhopadhyay C,Zhao X,Maroni D,Band V,Naramura M
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Fri Jul 11 00:00:00 EDT 2014
Highly upregulated Lhx2 in the Foxn1-/- nude mouse phenotype reflects a dysregulated and expanded epidermal stem cell niche.
PloS one
Bohr S,Patel SJ,Vasko R,Shen K,Huang G,Yarmush ML,Berthiaume F
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Mon Dec 23 00:00:00 EST 2013
The CD9/CD81 tetraspanin complex and tetraspanin CD151 regulate α3β1 integrin-dependent tumor cell behaviors by overlapping but distinct mechanisms.
PloS one
Gustafson-Wagner E,Stipp CS
14-0495 was used in Blocking experiments to study the effects of tetraspanin interactions on the pro-migratory functions of integrin α3β1.
Mon Nov 11 00:00:00 EST 2013
Stem-like epithelial cells are concentrated in the distal end of the fallopian tube: a site for injury and serous cancer initiation.
Stem cells (Dayton, Ohio)
Paik DY,Janzen DM,Schafenacker AM,Velasco VS,Shung MS,Cheng D,Huang J,Witte ON,Memarzadeh S
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 14-0495-82) in Flow Cytometry
Thu Nov 01 00:00:00 EDT 2012
P2Y2 receptor inhibits EGF-induced MAPK pathway to stabilise keratinocyte hemidesmosomes.
Journal of cell science
Faure E,Garrouste F,Parat F,Monferran S,Leloup L,Pommier G,Kovacic H,Lehmann M
14-0495 was used in Immunofluorescence to show the role of P2Y2R and Gαq in ERK1/2 signalling inhibition, modulation of hemidesmosome dynamics and keratinocyte migration.
Sat Sep 15 00:00:00 EDT 2012
The interplay between surfaces and soluble factors define the immunologic and angiogenic properties of myeloid dendritic cells.
BMC immunology
Sprague L,Muccioli M,Pate M,Meles E,McGinty J,Nandigam H,Venkatesh AK,Gu MY,Mansfield K,Rutowski A,Omosebi O,Courreges MC,Benencia F
14-0495 was used in Flow cytometry/Cell sorting to study the effects of different surfaces and soluble factors on the biology of dendritic cells.
Mon Jun 06 00:00:00 EDT 2011
Hepatoblast-like progenitor cells derived from embryonic stem cells can repopulate livers of mice.
Gastroenterology
Li F,Liu P,Liu C,Xiang D,Deng L,Li W,Wangensteen K,Song J,Ma Y,Hui L,Wei L,Li L,Ding X,Hu Y,He Z,Wang X
14-0495 was used in Flow cytometry/Cell sorting to identify and purify ES cell-derived hepatoblast-like progenitor cells and to explore their capacity for liver repopulation in mice after in vitro expansion.
Wed Dec 01 00:00:00 EST 2010