Product References
Stemness properties of SSEA-4+ subpopulation isolated from heterogenous Wharton's jelly mesenchymal stem/stromal cells.
Frontiers in cell and developmental biology
Smolinska A,Chodkowska M,Kominek A,Janiec J,Piwocka K,Sulejczak D,Sarnowska A
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Sat Mar 09 00:00:00 UTC 2024
Isolation and characterization mesenchymal stem cells from red panda (Ailurus fulgens styani) endometrium.
Conservation physiology
Wang DH,Wu XM,Chen JS,Cai ZG,An JH,Zhang MY,Li Y,Li FP,Hou R,Liu YL
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Sat Jan 01 00:00:00 UTC 2022
Tumor suppressor DEAR1 regulates mammary epithelial cell fate and predicts early onset and metastasis in triple negative breast cancer.
Scientific reports
Le UQ,Chen N,Balasenthil S,Lurie E,Yang F,Liu S,Rubin L,Solis Soto LM,Raso MG,Batra H,Sahin AA,Wistuba II,Killary AM
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Mon Nov 14 00:00:00 UTC 2022
Ablating Lgr5-expressing prostatic stromal cells activates the ERK-mediated mechanosensory signaling and disrupts prostate tissue homeostasis.
Cell reports
Wei X,Zhang L,Zhang Y,Cooper C,Brewer C,Tsai CF,Wang YT,Glaz M,Wessells HB,Que J,Titus MA,Cirulli V,Glaser A,Liu T,Reder NP,Creighton CJ,Xin L
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Tue Sep 06 00:00:00 UTC 2022
The Effect of Infant Gastric Digestion on Human Maternal Milk Cells.
Molecular nutrition & food research
Doerfler R,Melamed JR,Whitehead KA
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Sat Oct 01 00:00:00 UTC 2022
ROR2 regulates self-renewal and maintenance of hair follicle stem cells.
Nature communications
Veltri A,Lang CMR,Cangiotti G,Chan CK,Lien WH
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2022
Mechanism of integrin activation by talin and its cooperation with kindlin.
Nature communications
Lu F,Zhu L,Bromberger T,Yang J,Yang Q,Liu J,Plow EF,Moser M,Qin J
17-0495-82 was used in Flow cytometry/Cell sorting to show that while being capable of inducing integrin activation, talin-H alone exhibits unexpectedly low potency versus a constitutively activated full-length talin.
Fri Apr 29 00:00:00 UTC 2022
Endogenous retroviruses promote homeostatic and inflammatory responses to the microbiota.
Cell
Lima-Junior DS,Krishnamurthy SR,Bouladoux N,Collins N,Han SJ,Chen EY,Constantinides MG,Link VM,Lim AI,Enamorado M,Cataisson C,Gil L,Rao I,Farley TK,Koroleva G,Attig J,Yuspa SH,Fischbach MA,Kassiotis G,Belkaid Y
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Thu Jul 08 00:00:00 UTC 2021
CBP-mediated Wnt3a/β-catenin signaling promotes cervical oncogenesis initiated by Piwil2.
Neoplasia (New York, N.Y.)
Feng D,Yan K,Liang H,Liang J,Wang W,Yu H,Zhou Y,Zhao W,Dong Z,Ling B
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
GRHL3 activates FSCN1 to relax cell-cell adhesions between migrating keratinocytes during wound reepithelialization.
JCI insight
Kashgari G,Venkatesh S,Refuerzo S,Pham B,Bayat A,Klein RH,Ramos R,Ta AP,Plikus MV,Wang PH,Andersen B
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Wed Sep 08 00:00:00 UTC 2021
Single-cell analyses unravel cell type-specific responses to a vitamin D analog in prostatic precancerous lesions.
Science advances
Abu El Maaty MA,Grelet E,Keime C,Rerra AI,Gantzer J,Emprou C,Terzic J,Lutzing R,Bornert JM,Laverny G,Metzger D
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Thu Jul 01 00:00:00 UTC 2021
Diverse epithelial cell populations contribute to the regeneration of secretory units in injured salivary glands.
Development (Cambridge, England)
Ninche N,Kwak M,Ghazizadeh S
17-0495-82 was used in Flow Cytometry to show that de novo formation of acini involves induction of cellular plasticity in multiple non-acinar cell populations.
Fri Oct 09 00:00:00 UTC 2020
An IFT20 mechanotrafficking axis is required for integrin recycling, focal adhesion dynamics, and polarized cell migration.
Molecular biology of the cell
Su S,Begum S,Ezratty EJ
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Sat Aug 01 00:00:00 UTC 2020
Chronic expression of p16INK4a in the epidermis induces Wnt-mediated hyperplasia and promotes tumor initiation.
Nature communications
Azazmeh N,Assouline B,Winter E,Ruppo S,Nevo Y,Maly A,Meir K,Witkiewicz AK,Cohen J,Rizou SV,Pikarsky E,Luxenburg C,Gorgoulis VG,Ben-Porath I
17-0495 was used in Flow cytometry/Cell sorting to investigate the effect of prolonged expression of transgenic p16INK4a in the mouse epidermis and the pathways involved in this effect.
Mon Jun 01 00:00:00 UTC 2020
Chronic expression of p16INK4a in the epidermis induces Wnt-mediated hyperplasia and promotes tumor initiation.
Nature communications
Azazmeh N,Assouline B,Winter E,Ruppo S,Nevo Y,Maly A,Meir K,Witkiewicz AK,Cohen J,Rizou SV,Pikarsky E,Luxenburg C,Gorgoulis VG,Ben-Porath I
17-0495 was used in Flow cytometry/Cell sorting to investigate the effect of prolonged expression of transgenic p16INK4a in the mouse epidermis and the pathways involved in this effect.
Mon Jun 01 00:00:00 UTC 2020
Chemotherapeutic Stress Influences Epithelial-Mesenchymal Transition and Stemness in Cancer Stem Cells of Triple-Negative Breast Cancer.
International journal of molecular sciences
Li X,Strietz J,Bleilevens A,Stickeler E,Maurer J
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Wed Jan 08 00:00:00 UTC 2020
Polyisocyanide Hydrogels as a Tunable Platform for Mammary Gland Organoid Formation.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Tang C,Span PN,Rowan AE,Aalders TW,Schalken JA,Adema GJ,Kouwer PHJ,Zegers MMP,Ansems M
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2020
Polyisocyanide Hydrogels as a Tunable Platform for Mammary Gland Organoid Formation.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Tang C,Span PN,Rowan AE,Aalders TW,Schalken JA,Adema GJ,Kouwer PHJ,Zegers MMP,Ansems M
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2020
Discrete populations of isotype-switched memory B lymphocytes are maintained in murine spleen and bone marrow.
Nature communications
Riedel R,Addo R,Ferreira-Gomes M,Heinz GA,Heinrich F,Kummer J,Greiff V,Schulz D,Klaeden C,Cornelis R,Menzel U,Kröger S,Stervbo U,Köhler R,Haftmann C,Kühnel S,Lehmann K,Maschmeyer P,McGrath M,Naundorf S,Hahne S,Sercan-Alp Ö,Siracusa F,Stefanowski J,Weber M,Westendorf K,Zimmermann J,Hauser AE,Reddy ST,Durek P,Chang HD,Mashreghi MF,Radbruch A
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Fri May 22 00:00:00 UTC 2020
A Sox2-Sox9 signalling axis maintains human breast luminal progenitor and breast cancer stem cells.
Oncogene
Domenici G,Aurrekoetxea-Rodríguez I,Simões BM,Rábano M,Lee SY,Millán JS,Comaills V,Oliemuller E,López-Ruiz JA,Zabalza I,Howard BA,Kypta RM,Vivanco MD
17-0495 was used in Flow cytometry/Cell sorting to reveal that Sox2 increases and estrogen reduces the expression of the transcription factor Sox9 which highlights the relevance of the SOX family transcription factors as therapeutic targets in breast cancer.
Mon Apr 01 00:00:00 UTC 2019
The MMTV-Wnt1 murine model produces two phenotypically distinct subtypes of mammary tumors with unique therapeutic responses to an EGFR inhibitor.
Disease models & mechanisms
Pfefferle AD,Darr DB,Calhoun BC,Mott KR,Rosen JM,Perou CM
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Fri Jul 05 00:00:00 UTC 2019
A basal-enriched microRNA is required for prostate tumorigenesis in a Pten knockout mouse model.
Molecular carcinogenesis
Fan X,Bjerke GA,Riemondy K,Wang L,Yi R
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Sun Dec 01 00:00:00 UTC 2019
Regulatory T Cells Control the Switch From in situ to Invasive Breast Cancer.
Frontiers in immunology
Martinez LM,Robila V,Clark NM,Du W,Idowu MO,Rutkowski MR,Bos PD
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Tue Sep 29 00:00:00 UTC 2020
Autophagy inhibition elicits emergence from metastatic dormancy by inducing and stabilizing Pfkfb3 expression.
Nature communications
La Belle Flynn A,Calhoun BC,Sharma A,Chang JC,Almasan A,Schiemann WP
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Wed Aug 14 00:00:00 UTC 2019
Isolation and Culture of Human Stem Cells from Apical Papilla under Low Oxygen Concentration Highlight Original Properties.
Cells
Rémy M,Ferraro F,Le Salver P,Rey S,Genot E,Djavaheri-Mergny M,Thébaud N,Boiziau C,Boeuf H
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Thu Nov 21 00:00:00 UTC 2019
Quiescent Tissue Stem Cells Evade Immune Surveillance.
Immunity
Agudo J,Park ES,Rose SA,Alibo E,Sweeney R,Dhainaut M,Kobayashi KS,Sachidanandam R,Baccarini A,Merad M,Brown BD
17-0495 was used in Flow cytometry/Cell sorting to identify a link between stem cell quiescence, antigen presentation, and immune evasion.
Tue Feb 20 00:00:00 UTC 2018
Integrin-Rac signalling for mammary epithelial stem cell self-renewal.
Breast cancer research : BCR
Olabi S,Ucar A,Brennan K,Streuli CH
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Mon Oct 22 00:00:00 UTC 2018
The hypoxic tumor microenvironment in vivo selects the cancer stem cell fate of breast cancer cells.
Breast cancer research : BCR
Kim H,Lin Q,Glazer PM,Yun Z
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Tue Mar 06 00:00:00 UTC 2018
Profiling human breast epithelial cells using single cell RNA sequencing identifies cell diversity.
Nature communications
Nguyen QH,Pervolarakis N,Blake K,Ma D,Davis RT,James N,Phung AT,Willey E,Kumar R,Jabart E,Driver I,Rock J,Goga A,Khan SA,Lawson DA,Werb Z,Kessenbrock K
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Wed May 23 00:00:00 UTC 2018
Single-Cell Analysis Identifies LY6D as a Marker Linking Castration-Resistant Prostate Luminal Cells to Prostate Progenitors and Cancer.
Cell reports
Barros-Silva JD,Linn DE,Steiner I,Guo G,Ali A,Pakula H,Ashton G,Peset I,Brown M,Clarke NW,Bronson RT,Yuan GC,Orkin SH,Li Z,Baena E
17-0495 was used in Flow cytometry/Cell sorting to analyse the molecular heterogeneity in basal and luminal prostate compartments.
Tue Dec 18 00:00:00 UTC 2018
Single-Cell Analysis Identifies LY6D as a Marker Linking Castration-Resistant Prostate Luminal Cells to Prostate Progenitors and Cancer.
Cell reports
Barros-Silva JD,Linn DE,Steiner I,Guo G,Ali A,Pakula H,Ashton G,Peset I,Brown M,Clarke NW,Bronson RT,Yuan GC,Orkin SH,Li Z,Baena E
17-0495 was used in Flow cytometry/Cell sorting to analyse the molecular heterogeneity in basal and luminal prostate compartments.
Tue Dec 18 00:00:00 UTC 2018
Epigenome-wide SRC-1-Mediated Gene Silencing Represses Cellular Differentiation in Advanced Breast Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research
Ward E,Varešlija D,Charmsaz S,Fagan A,Browne AL,Cosgrove N,Cocchiglia S,Purcell SP,Hudson L,Das S,O'Connor D,O'Halloran PJ,Sims AH,Hill AD,Young LS
17-0495-82 was used in Flow Cytometry to reveal an epigenetic reprograming pathway, whereby concerted differential DNA methylation is potentiated by SRC-1 in the endocrine resistant setting.
Wed Aug 01 00:00:00 UTC 2018
Inflammatory memory sensitizes skin epithelial stem cells to tissue damage.
Nature
Naik S,Larsen SB,Gomez NC,Alaverdyan K,Sendoel A,Yuan S,Polak L,Kulukian A,Chai S,Fuchs E
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Thu Oct 26 00:00:00 UTC 2017
Autoreactive T Cells and Chronic Fungal Infection Drive Esophageal Carcinogenesis.
Cell host & microbe
Zhu F,Willette-Brown J,Song NY,Lomada D,Song Y,Xue L,Gray Z,Zhao Z,Davis SR,Sun Z,Zhang P,Wu X,Zhan Q,Richie ER,Hu Y
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Wed Apr 12 00:00:00 UTC 2017
Low Testosterone Alters the Activity of Mouse Prostate Stem Cells.
The Prostate
Zhou Y,Copeland B,Otto-Duessel M,He M,Markel S,Synold TW,Jones JO
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Sat Apr 01 00:00:00 UTC 2017
Induced p53 loss in mouse luminal cells causes clonal expansion and development of mammary tumours.
Nature communications
Tao L,Xiang D,Xie Y,Bronson RT,Li Z
17-0495 was used in Flow cytometry/Cell sorting to investigate how p53 deficiency contributes to breast tumourigenesis from luminal cells.
Mon Feb 13 00:00:00 UTC 2017
Induced p53 loss in mouse luminal cells causes clonal expansion and development of mammary tumours.
Nature communications
Tao L,Xiang D,Xie Y,Bronson RT,Li Z
17-0495 was used in Flow cytometry/Cell sorting to investigate how p53 deficiency contributes to breast tumourigenesis from luminal cells.
Mon Feb 13 00:00:00 UTC 2017
Characterization of the CD49f+/CD44+/CD24- single-cell derived stem cell population in basal-like DCIS cells.
Oncotarget
Duru N,Gernapudi R,Lo PK,Yao Y,Wolfson B,Zhang Y,Zhou Q
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Tue Jul 26 00:00:00 UTC 2016
Inhibition of the glucocorticoid receptor results in an enhanced miR-99a/100-mediated radiation response in stem-like cells from human prostate cancers.
Oncotarget
Rane JK,Erb HH,Nappo G,Mann VM,Simms MS,Collins AT,Visakorpi T,Maitland NJ
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Tue Aug 09 00:00:00 UTC 2016
Directed elimination of senescent cells by inhibition of BCL-W and BCL-XL.
Nature communications
Yosef R,Pilpel N,Tokarsky-Amiel R,Biran A,Ovadya Y,Cohen S,Vadai E,Dassa L,Shahar E,Condiotti R,Ben-Porath I,Krizhanovsky V
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Wed Apr 06 00:00:00 UTC 2016
Yap1 promotes the survival and self-renewal of breast tumor initiating cells via inhibiting Smad3 signaling.
Oncotarget
Sun JG,Chen XW,Zhang LP,Wang J,Diehn M
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Tue Mar 01 00:00:00 UTC 2016
Llgl1 prevents metaplastic survival driven by epidermal growth factor dependent migration.
Oncotarget
Greenwood E,Maisel S,Ebertz D,Russ A,Pandey R,Schroeder J
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Tue Sep 20 00:00:00 UTC 2016
Analysis of histone H2BGFP retention in mouse submandibular gland reveals actively dividing stem cell populations.
Stem cells and development
Kwak M,Ghazizadeh S
17-0495 was used in Flow cytometry/Cell sorting to study the population dynamics in the salivary gland, and to identify the quiescent stem cells residing in the gland.
Sun Mar 01 00:00:00 UTC 2015
Enrichment of Human Stem-Like Prostate Cells with s-SHIP Promoter Activity Uncovers a Role in Stemness for the Long Noncoding RNA H19.
Stem cells and development
Bauderlique-Le Roy H,Vennin C,Brocqueville G,Spruyt N,Adriaenssens E,Bourette RP
17-0495 was used in Flow cytometry/Cell sorting to provide the first evidence that s-SHIP-GFP promoter reporter offers a unique marker for the enrichment of human stem-like cell populations.
Fri May 15 00:00:00 UTC 2015
The Transcriptional Repressor ZNF503/Zeppo2 Promotes Mammary Epithelial Cell Proliferation and Enhances Cell Invasion.
The Journal of biological chemistry
Shahi P,Slorach EM,Wang CY,Chou J,Lu A,Ruderisch A,Werb Z
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Fri Feb 06 00:00:00 UTC 2015
Merlin Isoforms 1 and 2 Both Act as Tumour Suppressors and Are Required for Optimal Sperm Maturation.
PloS one
Zoch A,Mayerl S,Schulz A,Greither T,Frappart L,Rübsam J,Heuer H,Giovannini M,Morrison H
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Mon May 23 00:00:00 UTC 2016
Distinct breast cancer stem/progenitor cell populations require either HIF1α or loss of PHD3 to expand under hypoxic conditions.
Oncotarget
Iriondo O,Rábano M,Domenici G,Carlevaris O,López-Ruiz JA,Zabalza I,Berra E,Vivanco Md
17-0495 was used in Flow cytometry/Cell sorting to show that hypoxic conditions expand cancer stem cell populations through distinct molecular mechanisms.
Tue Oct 13 00:00:00 UTC 2015
NDY1/KDM2B functions as a master regulator of polycomb complexes and controls self-renewal of breast cancer stem cells.
Cancer research
Kottakis F,Foltopoulou P,Sanidas I,Keller P,Wronski A,Dake BT,Ezell SA,Shen Z,Naber SP,Hinds PW,McNiel E,Kuperwasser C,Tsichlis PN
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Tue Jul 15 00:00:00 UTC 2014
FGF ligands of the postnatal mammary stroma regulate distinct aspects of epithelial morphogenesis.
Development (Cambridge, England)
Zhang X,Martinez D,Koledova Z,Qiao G,Streuli CH,Lu P
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Mon Sep 01 00:00:00 UTC 2014
Targeting unique metabolic properties of breast tumor initiating cells.
Stem cells (Dayton, Ohio)
Feng W,Gentles A,Nair RV,Huang M,Lin Y,Lee CY,Cai S,Scheeren FA,Kuo AH,Diehn M
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Tue Jul 01 00:00:00 UTC 2014
NRF2/long noncoding RNA ROR signaling regulates mammary stem cell expansion and protects against estrogen genotoxicity.
The Journal of biological chemistry
Zhang Y,Xia J,Li Q,Yao Y,Eades G,Gernapudi R,Duru N,Kensler TW,Zhou Q
17-0495 was used in Flow cytometry/Cell sorting to provide further evidence for the critical role of NRF2 in maintaining normal stem cell subpopulations in mammary epithelium.
Fri Nov 07 00:00:00 UTC 2014
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
17-0495 was used in Flow cytometry/Cell sorting to study how the GATA3-miR-29b axis regulates the tumour microenvironment, and inhibits metastasis to open up possibilities for therapeutic intervention in breast cancer.
Fri Feb 01 00:00:00 UTC 2013
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
17-0495 was used in Flow cytometry/Cell sorting to study how the GATA3-miR-29b axis regulates the tumour microenvironment, and inhibits metastasis to open up possibilities for therapeutic intervention in breast cancer.
Fri Feb 01 00:00:00 UTC 2013
Evidence for a multipotent mammary progenitor with pregnancy-specific activity.
Breast cancer research : BCR
Kaanta AS,Virtanen C,Selfors LM,Brugge JS,Neel BG
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Mon Dec 08 00:00:00 UTC 2014
Lgr4 is a key regulator of prostate development and prostate stem cell differentiation.
Stem cells (Dayton, Ohio)
Luo W,Rodriguez M,Valdez JM,Zhu X,Tan K,Li D,Siwko S,Xin L,Liu M
17-0495 was used in Flow cytometry/Cell sorting to investigate mechanisms modulating prostate cell fate determination, showing that Lgr4 is a key regulator of development.
Fri Nov 01 00:00:00 UTC 2013
Generation of a novel germline stem cell line expressing a germline-specific reporter in the mouse.
Genesis (New York, N.Y. : 2000)
Shiura H,Ikeda R,Lee J,Sato T,Ogonuki N,Hirose M,Ogura A,Ogawa T,Abe K
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Mon Jul 01 00:00:00 UTC 2013
Distinct effects of EGFR ligands on human mammary epithelial cell differentiation.
PloS one
Mukhopadhyay C,Zhao X,Maroni D,Band V,Naramura M
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Fri Jul 11 00:00:00 UTC 2014
Highly upregulated Lhx2 in the Foxn1-/- nude mouse phenotype reflects a dysregulated and expanded epidermal stem cell niche.
PloS one
Bohr S,Patel SJ,Vasko R,Shen K,Huang G,Yarmush ML,Berthiaume F
Published figure using CD49f (Integrin alpha 6) monoclonal antibody (Product # 17-0495-82) in Flow Cytometry
Mon Dec 23 00:00:00 UTC 2013
Interfollicular epidermal stem cells: boosting and rescuing from adult skin.
Methods in molecular biology (Clifton, N.J.)
Cerqueira MT,Frias AM,Reis RL,Marques AP
17-0495-82 was used in Immunomagnetic Selection to described indulges the boosting and consecutive purification and separation of interfollicular epidermal stem cells from human keratinocytes cultures.
Wed Sep 04 00:00:00 UTC 2013
Leptin deficiency suppresses MMTV-Wnt-1 mammary tumor growth in obese mice and abrogates tumor initiating cell survival.
Endocrine-related cancer
Zheng Q,Dunlap SM,Zhu J,Downs-Kelly E,Rich J,Hursting SD,Berger NA,Reizes O
17-0495 was used in Flow cytometry/Cell sorting to study how leptin deficiency can suppress both tumour growth and tumour initiating cell survival in obese mice.
Mon Aug 01 00:00:00 UTC 2011
Novel stem/progenitor cell population from murine tracheal submucosal gland ducts with multipotent regenerative potential.
Stem cells (Dayton, Ohio)
Hegab AE,Ha VL,Gilbert JL,Zhang KX,Malkoski SP,Chon AT,Darmawan DO,Bisht B,Ooi AT,Pellegrini M,Nickerson DW,Gomperts BN
17-0495 was used in Flow cytometry/Cell sorting to demonstrate that submucosal gland duct cells are a multipotent stem cell for airway epithelial repair.
Mon Aug 01 00:00:00 UTC 2011