Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QTGLSGQND TSQTSSPSAS SSMSGGIFLF FVANAIIHLF CFS (20-61 aa encoded by BC000644)
CD52, also known as CAMPATH-1 antigen, is a glycoprotein that is expressed on the surface of various immune cells, including T and B lymphocytes, natural killer cells, and monocytes. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including multiple sclerosis, chronic lymphocytic leukemia, and autoimmune disorders. CD52 is the target of alemtuzumab, a monoclonal antibody that is used to treat these conditions by depleting immune cells that express CD52.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cambridge pathology 1 antigen; CAMPATH-1 antigen; CD52; CD52 antigen; CD52 antigen (CAMPATH-1 antigen); CDw52; CDW52 antigen (CAMPATH-1 antigen); Epididymal secretory protein E5; epididymis secretory sperm binding protein Li 171mP; He5; HEL-S-171mP; Human epididymis-specific protein 5
Gene Aliases: CD52; CDW52; HE5
UniProt ID: (Human) P31358
Entrez Gene ID: (Human) 1043
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support