Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Does not react with Baboon or Horse
CD53 is a tetraspanin family transmembrane glycoprotein expressed in the lymphoid-myeloid lineage. This molecule has been reported to form complexes with other leukocyte surface proteins such as CD2, CD19, CD21, MHC II, VLA-4 or tetraspanins CD37, CD81 and CD82, thus probably modulating various signaling processes. CD53 is involved in radioresistancy of tumor cells and its triggering has anti-apoptotic effect. In thymus, CD53 is up-regulated in response to positive selection signals during T cell development, and is strongly expressed upon macrophage exposure to bacterial lipopolysaccharide, whereas stimulation of neutrophils results in down-regulation of CD53 expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen MOX44 identified by monoclonal antibody MRC-OX44; CD53; CD53 antigen; CD53 glycoprotein; CD53 tetraspan antigen; cell surface antigen; Cell surface glycoprotein CD53; Leukocyte surface antigen CD53; Tetraspanin-25; transmembrane glycoprotein; Tspan-25
Gene Aliases: CD53; MOX44; TSPAN25
UniProt ID: (Human) P19397
Entrez Gene ID: (Human) 963
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support