Product References
A Matrix-Assisted Laser Desorption/Ionization Time-of-Flight Assay Identifies Nilotinib as an Inhibitor of Inflammation in Acute Myeloid Leukemia.
Journal of medicinal chemistry
Marín-Rubio JL,Peltier-Heap RE,Dueñas ME,Heunis T,Dannoura A,Inns J,Scott J,Simpson AJ,Blair HJ,Heidenreich O,Allan JM,Watt JE,Martin MP,Saxty B,Trost M
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Flow Cytometry
Thu Sep 22 00:00:00 UTC 2022
Macrophage ICAM-1 functions as a regulator of phagocytosis in LPS induced endotoxemia.
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Zhong H,Lin H,Pang Q,Zhuang J,Liu X,Li X,Liu J,Tang J
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Neutralization
Mon Feb 01 00:00:00 UTC 2021
Macrophage ICAM-1 functions as a regulator of phagocytosis in LPS induced endotoxemia.
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Zhong H,Lin H,Pang Q,Zhuang J,Liu X,Li X,Liu J,Tang J
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Neutralization
Mon Feb 01 00:00:00 UTC 2021
ICAM-1 and ICAM-2 Are Differentially Expressed and Up-Regulated on Inflamed Pulmonary Epithelium, but Neither ICAM-2 nor LFA-1: ICAM-1 Are Required for Neutrophil Migration Into the Airways In Vivo.
Frontiers in immunology
Chong DLW,Rebeyrol C,José RJ,Williams AE,Brown JS,Scotton CJ,Porter JC
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Immunohistochemistry
Mon Oct 25 00:00:00 UTC 2021
Conserved regulation of RNA processing in somatic cell reprogramming.
BMC genomics
Kanitz A,Syed AP,Kaji K,Zavolan M
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Flow Cytometry
Thu Jan 31 00:00:00 UTC 2019
VCAM-1 Density and Tumor Perfusion Predict T-cell Infiltration and Treatment Response in Preclinical Models.
Neoplasia (New York, N.Y.)
Riegler J,Gill H,Ogasawara A,Hedehus M,Javinal V,Oeh J,Ferl GZ,Marik J,Williams S,Sampath D,Schartner J,Carano RAD
16-0541 was used in Flow cytometry/Cell sorting to test if non-invasive imaging of tumor perfusion and vascular inflammation may be able to explain differences in T-cell infiltration in pre-clinical tumor models, relevant for treatment outcomes.
Tue Oct 01 00:00:00 UTC 2019
PAI1 mediates fibroblast-mast cell interactions in skin fibrosis.
The Journal of clinical investigation
Pincha N,Hajam EY,Badarinath K,Batta SPR,Masudi T,Dey R,Andreasen P,Kawakami T,Samuel R,George R,Danda D,Jacob PM,Jamora C
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Neutralization
Tue May 01 00:00:00 UTC 2018
Low-Dose Radiotherapy Has No Harmful Effects on Key Cells of Healthy Non-Inflamed Joints.
International journal of molecular sciences
Deloch L,Rückert M,Fietkau R,Frey B,Gaipl US
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Flow Cytometry
Tue Oct 16 00:00:00 UTC 2018
Fine-Tuning Mybl2 Is Required for Proper Mesenchymal-to-Epithelial Transition during Somatic Reprogramming.
Cell reports
Ward C,Volpe G,Cauchy P,Ptasinska A,Almaghrabi R,Blakemore D,Nafria M,Kestner D,Frampton J,Murphy G,Buganim Y,Kaji K,García P
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Flow Cytometry
Tue Aug 07 00:00:00 UTC 2018
Constitutively Active SMAD2/3 Are Broad-Scope Potentiators of Transcription-Factor-Mediated Cellular Reprogramming.
Cell stem cell
Ruetz T,Pfisterer U,Di Stefano B,Ashmore J,Beniazza M,Tian TV,Kaemena DF,Tosti L,Tan W,Manning JR,Chantzoura E,Ottosson DR,Collombet S,Johnsson A,Cohen E,Yusa K,Linnarsson S,Graf T,Parmar M,Kaji K
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Flow Cytometry
Thu Dec 07 00:00:00 UTC 2017
Fibroblastic reticular cells of the lymphoid tissues modulate T cell activation threshold during homeostasis via hyperactive cyclooxygenase-2/prostaglandin E2 axis.
Scientific reports
Yu M,Guo G,Zhang X,Li L,Yang W,Bollag R,Cui Y
16-0541 was used in Flow cytometry/Cell sorting to identify an intrinsic mechanism of fibroblastic reticular cells shared between mice and humans for suppressing T cell sensitivity to activation via PGE2.
Tue Jun 13 00:00:00 UTC 2017
Treg engage lymphotoxin beta receptor for afferent lymphatic transendothelial migration.
Nature communications
Brinkman CC,Iwami D,Hritzo MK,Xiong Y,Ahmad S,Simon T,Hippen KL,Blazar BR,Bromberg JS
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Flow Cytometry
Tue Jun 21 00:00:00 UTC 2016
Treg engage lymphotoxin beta receptor for afferent lymphatic transendothelial migration.
Nature communications
Brinkman CC,Iwami D,Hritzo MK,Xiong Y,Ahmad S,Simon T,Hippen KL,Blazar BR,Bromberg JS
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Flow Cytometry
Tue Jun 21 00:00:00 UTC 2016
Treg engage lymphotoxin beta receptor for afferent lymphatic transendothelial migration.
Nature communications
Brinkman CC,Iwami D,Hritzo MK,Xiong Y,Ahmad S,Simon T,Hippen KL,Blazar BR,Bromberg JS
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Flow Cytometry
Tue Jun 21 00:00:00 UTC 2016
Palmitoyl acyltransferase DHHC21 mediates endothelial dysfunction in systemic inflammatory response syndrome.
Nature communications
Beard RS,Yang X,Meegan JE,Overstreet JW,Yang CG,Elliott JA,Reynolds JJ,Cha BJ,Pivetti CD,Mitchell DA,Wu MH,Deschenes RJ,Yuan SY
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Immunofluorescence
Thu Sep 22 00:00:00 UTC 2016
Palmitoyl acyltransferase DHHC21 mediates endothelial dysfunction in systemic inflammatory response syndrome.
Nature communications
Beard RS,Yang X,Meegan JE,Overstreet JW,Yang CG,Elliott JA,Reynolds JJ,Cha BJ,Pivetti CD,Mitchell DA,Wu MH,Deschenes RJ,Yuan SY
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Immunofluorescence
Thu Sep 22 00:00:00 UTC 2016
Ectonucleotidase activity and immunosuppression in astrocyte-CD4 T cell bidirectional signaling.
Oncotarget
Filipello F,Pozzi D,Proietti M,Romagnani A,Mazzitelli S,Matteoli M,Verderio C,Grassi F
16-0541 was used in Flow cytometry/Cell sorting to investigate how TCR-independent interactions promote an immunosuppressive program in T cells to control possible brain injury by deregulated T cell activation during neuroinflammation.
Tue Feb 02 00:00:00 UTC 2016
Intercellular adhesion molecule-1 expression by skeletal muscle cells augments myogenesis.
Experimental cell research
Goh Q,Dearth CL,Corbett JT,Pierre P,Chadee DN,Pizza FX
16-0541 was used in Neutralization experiments to reveal mechanisms through which skeletal muscle cell expression of ICAM-1 augments the regenerative and hypertrophic processes of myogenesis.
Sun Feb 15 00:00:00 UTC 2015
Reprogramming Roadblocks Are System Dependent.
Stem cell reports
Chantzoura E,Skylaki S,Menendez S,Kim SI,Johnsson A,Linnarsson S,Woltjen K,Chambers I,Kaji K
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Flow Cytometry
Tue Sep 08 00:00:00 UTC 2015
Natural killer T cells are essential for the development of contact hypersensitivity in BALB/c mice.
The Journal of investigative dermatology
Shimizuhira C,Otsuka A,Honda T,Kitoh A,Egawa G,Nakajima S,Nakashima C,Watarai H,Miyachi Y,Kabashima K
16-0541 was used in Flow cytometry/Cell sorting to report the pivotal contribution of dendritic and natural killer T cell interactions to hypersensitivity.
Sat Nov 01 00:00:00 UTC 2014
Dietary gluten increases natural killer cell cytotoxicity and cytokine secretion.
European journal of immunology
Larsen J,Dall M,Antvorskov JC,Weile C,Engkilde K,Josefsen K,Buschard K
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Flow Cytometry
Wed Oct 01 00:00:00 UTC 2014
High-resolution analysis with novel cell-surface markers identifies routes to iPS cells.
Nature
O'Malley J,Skylaki S,Iwabuchi KA,Chantzoura E,Ruetz T,Johnsson A,Tomlinson SR,Linnarsson S,Kaji K
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 16-0541-81) in Flow Cytometry
Thu Jul 04 00:00:00 UTC 2013
ICAM-1-dependent homotypic aggregates regulate CD8 T cell effector function and differentiation during T cell activation.
Journal of immunology (Baltimore, Md. : 1950)
Zumwalde NA,Domae E,Mescher MF,Shimizu Y
16-0541 was used in Flow cytometry/Cell sorting to demonstrate that during CD8 T cell activation, the T cell clustering mechanism allows for continued proliferation of T cells, and regulates CD8 T cell effector function and differentiation.
Tue Oct 01 00:00:00 UTC 2013
Lung-derived mesenchymal stromal cell post-transplantation survival, persistence, paracrine expression, and repair of elastase-injured lung.
Stem cells and development
Hoffman AM,Paxson JA,Mazan MR,Davis AM,Tyagi S,Murthy S,Ingenito EP
16-0541 was used in Blocking experiments to examine how lung-derived MSCs differ from syngeneic bone marrow MSCs in vitro and after transplantation in a murine model of emphysema.
Sat Oct 01 00:00:00 UTC 2011
Crosstalk between CXCR4/stromal derived factor-1 and VLA-4/VCAM-1 pathways regulates neutrophil retention in the bone marrow.
Journal of immunology (Baltimore, Md. : 1950)
Petty JM,Lenox CC,Weiss DJ,Poynter ME,Suratt BT
16-0541 was used in Neutralization experiments to show that alpha(4)beta(1) integrin is expressed on murine marrow neutrophils, decreases with maturation and its blockade leads to the release of marrow neutrophils.
Thu Jan 01 00:00:00 UTC 2009
Targeted delivery of oligodeoxynucleotides to mouse lung endothelial cells in vitro and in vivo.
Molecular therapy : the journal of the American Society of Gene Therapy
Wilson A,Zhou W,Champion HC,Alber S,Tang ZL,Kennel S,Watkins S,Huang L,Pitt B,Li S
16-0541 was used in Immunohistochemistry-immunofluorescence to develop a novel lipidic vector that is highly efficient in targeted delivery of oligodeoxynucleotides to pulmonary endothelium.
Thu Sep 01 00:00:00 UTC 2005